|
I am new to RNAi work. Which should I choose, siRNA or shRNA? Both methods provide viable knockdown efficiency, to better help you choose please refer to the table below.
Choosing the Appropriate RNAi Effector for your Application
| |
siRNA |
shRNA, Transfected plasmid |
shRNA, Lentivirus-Delivered |
| Cell Lines (actual efficacy highly cell type specific) |
| Immortalized/Cancer |
XXX |
XXX |
XXX |
| (Embryonic) Stem |
X |
X |
XXX |
| Primary |
X |
X |
XXX |
| Screening |
| Arrayed, (single) |
XXX |
XX |
XXX |
| Pooled, multiple genes / genome |
- |
- |
XXX |
| Pooled, single gene |
XXX |
X |
X |
| in vivo (routes) |
| Intratumoral |
XXX |
XXX |
XXX |
| ex vivo, implantation |
X |
X |
XXX |
| Cranial |
X |
X |
XX |
| Peritineum |
X |
X |
XX |
| Muscle |
XX |
XX |
XX |
My gene of interest is very stable. Which RNAi format do I choose? Both methods provide viable knockdown efficiency, to better help you choose please refer to the table below.
RNAi effector Features
| |
siRNA |
shRNA, Transfected plasmid |
shRNA, Lentivirus-Delivered |
| Silencing Duration |
Transient (48hrs to 1wk) |
Transient (48hrs to 1wk), or Stable, long-term |
Stable, long-term |
| mRNA silenced |
6 - 24 hours after transfection |
24 - 48 hours after transfection |
48 hours after transduction |
| Protein silenced |
48 hours to 5 days post-transfection |
48 hours to 5 days post-transfection |
72 hours to 6 days post-transduction |
| Ease of Use |
Simple |
Moderate |
Simple |
| What comes Standard |
~10 nmoles, desalted |
1µg, TE buffer |
200 µl of 1 x 10^6 TU/ml |
| Custom Options |
Yes |
Yes |
Yes |
For additional questions regarding our siRNA and shRNA product lines, please select one of the links below:
shRNA FAQs
|