Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Functional Genomics & RNAi > shRNA > Custom Cloning
shRNA
Custom Cloning
Exclusive offer for custom shRNA!
Purchase a minimum of 3 lentiviral clones and receive special $500 (Euro 400/GBP 350) per clone pricing, corresponding glycerol stocks, and a free lentiviral control* with every 3-clone purchase.
*Customer has choice of one empty or non-target custom lentiviral control per every 3 clones purchased.
To place an order or for more details, contact your local sales representative or email us at MISSIONRNAi@sial.com. Promotion is not valid on web orders.
Start your RNAi experiments today! Let the MISSION team clone your shRNA into your vector of choice and then provide you with ready to use lentiviral particles.
esiFlex™ Woman
  • Select by methods other than puromycin, including neomycin (G418) and FACS via reporters
  • Confirm your lentiviral delivery through a fluorescent reporter
  • Customize your shRNA sequence
  • Eliminate tedious cloning and viral production
There may be times when classic drug selection for stable or semi-stable cell lines is not possible or desired. In addition, orthogonal methods of selection from puromycin may be required for the expression of multiple shRNAs or expression of shRNA concomitant with transgene expression.
Vector Backbone Expression Selection Method Alternative to CMV promoter
Constitutive Inducible Puromycin Neomycin (G418) Fluorescence
pLKO.1-puro (186 Kb PDF) X   X     X
pLKO.1-CMV-tGFP (179 Kb PDF) X       X  
pLKO.1-puro-CMV-tGFP (196 Kb PDF) X   X   X  
pLKO.1-CMV-Neo (30 Kb PDF) X        
pLKO.1-Neo (30 Kb PDF) X       X
pLKO.1-Neo-CMV-tGFP (193 Kb PDF) X     X X  
pLKO.1-puro-CMV-TagCFP™ (27 Kb PDF) X   X   X  
pLKO.1-puro-CMV-TagYFP™ (188 Kb PDF) X   X   X  
pLKO.1-puro-CMV-TagRFP™ (188 Kb PDF) X   X   X  
pLKO.1-puro-CMV-TagFP635™ (188 Kb PDF) X   X   X  
pLKO.1-puro-UbC-TurboGFP™ (193 Kb PDF) X   X   X X
pLKO.1-puro-UbC-TagFP635™ (195 Kb PDF) X   X   X X
pLKO-puro-IPTG-1xLacO (95 Kb PDF)   X X     X
pLKO-puro-IPTG-3xLacO (399 Kb PDF)   X X     X
For vector sequence and maps of the custom cloning vectors visit our vector maps page.
Order and Service Information
Our service includes: shRNA design, cloning, sequence verification, DNA quantitation and titer determination (when ordering lentivirus). A DNA order is typically shipped in 6 weeks and an order for lentivirus is shipped 8 weeks from the time the order is placed. To order either: contact your local sales representative, fill out our online request form, or email us at MISSIONRNAi@sial.com.
* Standard DNA orders receive a total of 1 µg of purified plasmid DNA in Tris-EDTA (TE) buffer at a concentration 20 ng/µl.
* Standard lentivirus orders receive 200 µl (4 x 50 µl aliquots) at a minimum titer of 1 x 106 TU/ml. We routinely deliver virus at titers higher than 1 x 107 TU/ml.
Selection by Fluorescent Proteins
Sigma® now offers vectors containing multiple fluorescent protein alternatives (TurboGFP, TagCFP, TagYFP, TagRFP, and TagFP635). With fluorescent proteins in the vector, FACS and fluorescence microscopy techniques allow for rapid estimation of transduction efficiency and automated, rapid selection of cell populations that stably express the shRNA. The TurboGFP and far red fluorescent protein (TagFP635) are available in multiple backbones. Selection by fluorescent protein expression provides an orthogonal method when cells already stably express puromycin resistance. Sigma's MISSION bio-production team will customize the vector with your favorite shRNA sequence. To order either: contact your local sales representative, fill out our online request form, or email us at MISSIONRNAi@sial.com.
Selection by Neomycin
For selection by a second class of antibiotics, we offer vectors containing the neomycin resistance gene. These two vectors are pLKO.1-Neo and pLKO.1-CMV-Neo. Stable shRNA integrants can be found by selecting in G418 (A1720). As with puromycin, a cytotoxicity profile for your particular cell line will need to be generated. As an example see the puromycin protocol, but use G418 instead of puromycin. As a reference, a final concentration of 1 mg/ml of G418 has been used for selection of shRNA constructs in A549 cells. To order either: contact your local sales representative, fill out our online request form, or email us at MISSIONRNAi@sial.com.
Alternatives to CMV Promoter
Each type of promoter for your selection markers will offer different expression levels with different stabilities in each cell line. Importantly, the CMV promoter, while yielding strong expression in many cell lines, may not function in certain cells. Customer feedback and internal Sigma research and development work indicates that the CMV promoter will not function well in leukocytes, and certain mouse cells. In addition, the literature indicates that these promoters do not function in stem cells, a critical flaw for many researchers.1,2 We currently offer vector backbones that use other promoters than CMV. In fact, the vector backbone for the entire TRC collection utilizes the hPGK promoter for the puromycin selection and the U6 promoter for the shRNA. For data and protocols of how to utilize this vector in hES cell and lymphocytes, visit our shRNA protocols page. In addition, we offer the pLKO.1-Neo vector that uses the hPGK promoter driving the neomycin resistance gene for selection by G418. For certain mouse cells, the ubiquitin promoter (UbC) can be a viable alternative. The pLKO.1-puro UbC-TurboGFP construct and the pLKO.1-puro UbC-TagFP635 were generated for these types of applications. To order either: contact your local sales representative, fill out our online request form, or email us at MISSIONRNAi@sial.com.
References
  1. Tang, F. C. et al. Stable suppression of gene expression in murine embryonic stem cells by RNAi directed from DNA vector-based short hairpin RNA. Stem Cells, 22, 93–9 (2004). Pubmed ID: 14688395
  2. Michibata, H. et al. Inhibition of mouse GPM6A expression leads to decreased differentiation of neurons derived from mouse embryonic stem cells. Stem Cells Dev., 17, 641–51 (2008). Pubmed ID: 18522499
back to top