| Day 0 |
| |
0.1 |
|
Determine the number of femurs needed we estimate approximately 1.5 million BMDM per femur by the end of the protocol. |
| |
0.2 |
|
Make an ice-cold solution 20 ml of MEM alpha and maintain on ice. |
| |
0.3 |
|
Pre-warm growth media to 37 °C. |
| |
0.4 |
|
Euthanize mice as per protocols approved at your institution. |
| |
0.5 |
|
Remove femurs from the mice and immediately place in ice cold MEM alpha. |
| |
0.6 |
|
Using a 10 cc needle and 30 ml syringe flush bone marrow from femurs into 60 mm Petri dish. |
| |
0.7 |
|
Gently pipette the bone marrow to disrupt. |
| |
0.8 |
|
Centrifuge at room temperature for 5 minutes at 1,000 RPM. |
| |
0.9 |
|
Remove supernatant without disrupting cell pellet. |
| |
0.10 |
|
Immediately add 20 ml of pre-warmed growth media and gently resuspend by pipetting. |
| |
0.11 |
|
Transfer to T-75 and incubate overnight in tissue culture incubator. |
| |
| Day 1 |
| |
1.1 |
|
Pre-warm growth media. |
| |
1.2 |
|
Transfer to sterile 50 ml conical. |
| |
1.3 |
|
Centrifuge at room temperature for 5 minutes at 1,000 RPM. |
| |
1.4 |
|
Resuspend pellet in 50 ml of growth media. |
| |
1.5 |
|
Transfer to T-150 |
| |
1.6 |
|
Return to tissue culture incubator. |
| |
| |
| Day 3 |
| |
3.1 |
|
Pre-warm growth media. |
| |
3.2 |
|
Replace media on cells |
| |
3.3 |
|
Return flask to tissue culture incubator. |
| |
| Day 5 |
| |
5.1 |
|
Pre-warm growth media. |
| |
5.2 |
|
Remove media from flask. |
| |
5.3 |
|
Add 10 ml of DPBS DI per flask and remove cells with gentle scraping. |
| |
5.4 |
|
Cells were counted. |
| |
5.5 |
|
Centrifuge at room temperature for 5 minutes at 1,000 RPM. |
| |
5.6 |
|
Resuspend cells in growth media at a concentration of 1x106 cells/ml |
| |
5.7 |
|
Add 100 μl of cells in media to each well required in a 96-well plate |
| |
| Day 6 |
| |
6.1 |
|
Thaw MISSION lentivirus |
| |
6.2 |
|
Calculate volume of virus (Vv) in μl per well for an MOI of 8. |
| |
6.3 |
|
Utilizing the following formula calculate the volume of DMEM (Dv) needed per well to bind virus to beads then add DMEM to empty tube. Dv=50-Vv-1.5. |
| |
6.4 |
|
Add 1.5 μl of ExpressMag beads (SHM03) per well to the DMEM. |
| |
6.5 |
|
Gently mix by pipetting |
| |
6.6 |
|
Add 50 μl of the mixture per well. |
| |
6.7 |
|
Place 96-well plate on supermagnetic plate for 15 minutes at room temperature. |
| |
6.8 |
|
Return cells to tissue culture incubator over night. |
| |
| Day 7 |
| |
7.1 |
|
Pre-warm selection medium. |
| |
7.2 |
|
Remove media from wells in 96-well plate. |
| |
7.3 |
|
Replace with 100 µl per well of selection medium. |
| |
7.4 |
|
Return to tissue culture incubator. |
| |
| Day 9 |
| |
9.1 |
|
Repeat 7.1 - 7.4. |
| |
| Day 11 |
| |
11.1 |
|
Perform desired assay. |