| Day 0 |
| |
0.1 |
|
Seed HEK293T cells (20,000 cells/well of 96-well plate) in complete DME media. |
| |
| Day 1 |
| |
1.1 |
|
Thaw the vial of Lentiviral Packaging Mix at room temperature and place on ice. |
| |
1.2 |
|
Put three empty polypropylene tubes on ice. |
| |
1.3 |
|
Add 1 µl of Lentiviral Packaging Mix per well to the first tube. |
| |
1.4 |
|
Add 15 µl of serum free DME and 0.6 µl transfection reagent, Fugene 6, per well into the second tube. |
| |
1.5 |
|
Add 0.1 µg/well of shRNA transfer vector into the third tube. |
| |
1.6 |
|
Combine all transfection cocktail components together in one polypropylene vial. |
| |
1.7 |
|
Mix gently by pipetting up and down. |
| |
1.8 |
|
Incubate at room temperature for 15 min. |
| |
1.9 |
|
Equally divide the mixture among the well to be transfected. |
| |
1.10 |
|
Incubate cells at 37 °C overnight in tissue culture incubator. |
| |
| Day 2 |
| |
2.1 |
|
16 hours post-transfection, gently remove media from the transfected cells (avoid disturbing cells) and replace with 100 µl of the pre-warmed complete media per well of 96-well plate. |
| |
2.2 |
|
Incubate cells in incubator (37 °C and 5% CO2) for an additional 24 hours. |
| |
| Day 3 |
| |
3.1 |
|
Double glove before proceeding to 3.2. |
| |
3.2 |
|
Gently remove supernatant, now containing virus, and place it in a collection plate. |
| |
3.3 |
|
Add fresh complete DME to the wells and incubate cells at 37 °C overnight. |
| |
3.4 |
|
Cover, and store collection plate at 4 °C. |
| |
| Day 4 |
| |
4.1 |
|
Double glove before proceeding to 4.2. |
| |
4.2 |
|
Remove second viral supernatant from wells and add it to the collection plate from the day before. |
| |
4.3 |
|
Determine titer by p24 ELISA. |
| |
4.4 |
|
Store virus tightly sealed at –80 °C. |