Life Science > Functional Genomics & RNAi > shRNA > Learning Center > MISSION Application Data > Lentiviral Particle Production
shRNA
Lentiviral Particle Production
High-throughput Lentiviral Particle Production
Fig. 1. Actual Titer Measurements of Lentiviral Particles Manufactured by Sigma. This graph presents a representative sample of individual MISSION lentiviral particle preparations titered by our QC team. Viral particles are prepared in 96-well format using state-of-the-art robotics and are titered using the p24 assay. Sigma's minimum QC specification for a single preparation is 1 x 106 TU/ml.
Contact Us
For questions about the library, pricing and quotes or other concerns, please e-mail us at: RNAi@sial.com.
MISSION is a trademark of Sigma-Aldrich. Label License.
TrakMates is a registered trademark of Matrix Technologies Corporation.
back to top
back to Application Data
shRNA Homepage
RNAi Homepage