Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
shRNA
pLKO 1 Stability
pLKO.1 Stability Assessed by Diagnostic Restriction Digestion
pLKO.1 Stability Assessed by Diagnostic Restriction Digestion
Digestion of pLKO.1 library constructs by PvuII produces three fragments of 3.8 kb, 2.5 kb, and 0.8 kb. PvuII digestions of 244 library clones in pLKO.1 were performed following ten rounds of sequential copying and regrowth. No evidence of recombination was observed. A representative digestion gel from a fraction of wells in a library plate from a 10th generation copy is shown.
Contact Us
For questions about the library, pricing and quotes or other concerns, please e-mail us at: RNAi@sial.com.
MISSION is a trademark of Sigma-Aldrich. Label License.
TrakMates is a registered trademark of Matrix Technologies Corporation.
back to top
back to Application Data
shRNA Homepage
RNAi Homepage