|
|||||
|
|||||
|
Bacterial Glycerol Stock (SHCLNG) Bacterial Glycerol Stock (SHCLNG) Download the SHCLNG Technical Bulletin (583 kb PDF) Purified Plasmid DNA Format (SHCLND) Download the SHCLND Technical Bulletin (561 kb PDF) Lentiviral Format (SHCLNV) Download the SHCLNV Technical Bulletin (124 kb PDF) Purified plasmid control vectors may be directly used in transient or stable gene silencing (puromycin selection). In addition, self-inactivating replication incompetent viral particles can be produced in packaging cells (HEK293T) by co-transfection with compatible packaging plasmids. Visit our controls webpage for more information on specific controls. Individual control vectors are provided as 10 µg of plasmid DNA in Tris-EDTA (TE) buffer at a concentration of 500 ng/µl. Download the MISSION Control Vectors Technical Bulletin (226 Kb PDF) MISSION Control Transduction Particles Sigma offers a variety of positive, negative, and transduction controls in lentiviral format. Additionally, our most popular controls are offered in high titer format. Visit our controls webpage for more information on specific controls. Standard titer control transduction particles are provided as 200 µl of 106 TU/ml (via p24 titering assay). High titer control transduction particles are provided as 10 x 20 µl aliquots of 109 TU/ml (via p24 titering assay). Download the MISSION Control Transduction Particles Technical Bulletin (231 Kb PDF) MISSION Lentiviral Packaging Mix (SHP001) The MISSION Lentiviral Packaging Mix is an optimized formulation of two plasmids expressing the key HIV packaging genes and a heterologous viral envelope gene. It is designed to be co-transfected along with a compatible lentiviral transfer vector in order to create high-titer pseudo-typed lentiviral particles used for downstream transduction applications. Download the MISSION Lentiviral Packaging Mix Technical Bulletin (286 Kb PDF) The 3' untranslated regions (3'UTRs) of mRNA transcripts control many aspects of post-transcriptional regulation of genes. 3'UTRs are involved in regulating transcript stability, translational efficiency, and localization in the cell. 3'UTRs are also the predominant target sequences for endogenous microRNAs (miRNA) or small interfering RNAs (siRNA). Sigma-Aldrich and SwitchGear Genomics have teamed up to offer you the MISSION 3'UTR Lenti GoClone collection, which is a genome-wide collection of cloned human 3'UTRs. Each viral vector expresses an optimized luciferase reporter gene (RenSP) fused to a 3’UTR sequence. Download the MISSION 3'UTR Lenti GoClone Technical Bulletin (154 Kb PDF) |
|||||
|
|
|||||