Sigma-Aldrich
Life Science > Functional Genomics & RNAi > shRNA > MISSION® shRNA Library
shRNA
MISSION® shRNA Library
Introducing the New MISSION shRNA Library, powered by TRC.
Do you want to perform RNAi in non-transfectable or non-dividing cells? Do you need efficient, long-term knockdown and require a robust, quality system? Sigma® Life Science and The RNAi Consortium (TRC) provide the MISSION shRNA Library for human and mouse genomes, developed at the Broad Institute, a joint venture between MIT and Harvard.
The MISSION shRNA Library is exclusive to Sigma Life Science and contains almost 250,000 clones including more than 80,000 validated clones. It combines all of the highly trusted TRC1 content plus an additional  88,000 clones targeting  4,000 new human genes and more than 5,000  new mouse genes!
Sigma-Aldrich® is both a collaborative member and supplier of the MISSION shRNA Library, the most comprehensive and thoroughly validated shRNA collection available.
  • The MISSION shRNA Library
  • MISSION shRNA Design
  • Lentiviral Production Expertise
  • Screening
  • Flexible Formats and Custom Options
Request a Quote or More Information
Search for shRNA Clones
Start Your shRNA Research Today
Why experiment witht the old TRC library when you can get the bigger and better MISSION shRNA Library? The MISSION shRNA Library is exclusive to Sigma® Life Science and contains almost 250,000 clones including more than 80,000 validated clones. It combines all of the highly trusted TRC1 content plus an additional 88,000 clones targeting 4,000 new human genes and more than 5,000 new mouse genes: The new MISSION shRNA library provides unparalleled genome coverage and is available only at Sigma Life Science. Start your research today with the largest and most comprehensive genome-wide shRNA library on the market!
shRNA Biorevolution
Optimal Lentiviral-Based RNAi
  • Stable or transient transfection
  • Viral particles for transduction
  • Integrates for long-term knockdown
  • Transduces virtually any cell type (dividing or non-dividing)
  • No interferon response
  • Lack of recombination issues
  • Standard preparation of lentivirus yields a high titer of at least106 TU/ml
For introductory information on working with shRNA and/or Lentivirus, visit: Getting Started with shRNA
  MISSION shRNA Library          vs.                 TRC1 Library          
  Total Validated Total Validated
Human Genes 20,018 8,897 16,019 7,405
Mouse Genes 21,171 8,345 15,960 6,506
Total Genes 41,189 17,242 31,979 13,911
Human Clones 129,695 43,470 82,017 24,550
Mouse Clones 118,062 36,654 77,819 20,564
Total Clones 247,757 80,124 159,836 45,114
shRNA Library Info Human Graph
shRNA Library Info Mouse Graph
back to top
MISSION shRNA Design
MISSION shRNA clones, designed and developed by TRC at the Broad Institute of MIT and Harvard, include hairpin sequences comprised of a 21 base stem and a 6 base loop. Rules based design consisting of sequence, specificity, and position scoring are utilized to generate optimal shRNA sequences. The hairpin sequences are each cloned into the pLKO.1 vector and sequence verified. A minimum of 3-5 shRNA constructs are created for each target gene to provide varying levels of knockdown and to target different regions of mRNA transcript. Sigma-Aldrich guarantees that upon purchase of the defined clone set quantity on the shRNA detail page for your gene of interest (this varies, but is typically 5 clones), that at least one of those clones for a gene should yield greater than 70% knockdown. For a given RefSeq, there is often a shRNA clone targeting the 3'UTR for use in phenotypic rescue studies using cDNA expression constructs.
shRNA Designed for High Performance
  • Sequence, specificity & position scoring with the Broad Institute algorithm
  • Comprehensive target coverage with ~5 clones per gene
  • 3' UTR shRNA constructs for phenotypic rescue studies
Validation-Graph-small
Click on image to enlarge. (281 Kb PDF)
Figure 1. High-throughput validation of MISSION-TRC shRNA lentiviral transduction particles. Gene silencing effect of 60 different genes was measured at the mRNA transcript level and compared to pLKO.1-puro empty control particles. For each target gene, approximately 70% or greater knockdown was achieved.
back to top
Lentiviral Production Expertise
Sigma-Aldrich is committed to providing unmatched infrastructure and service for RNAi. With the addition of state-of-the-art robotics, liquid handling, LIMS, and dedicated laboratories in our new Life Science & High Technology Center, our goal is to provide superior quality and convenient service for your RNAi research needs.
High Quality Manufacturing Systems and Personnel
  • High Throughput (HT) robotic handling
  • 1D and 2D barcoding
  • Highly trained staff of R&D, Production, QC & QA Professionals
  • HT viral Production
  • High Content Screening
The Leader in Lentiviral shRNA Products and Support
  • Extensive web-based resources
  • Technical, R&D and Bioinformatics experts on staff
back to top
Screening Formats for Any Scale
Sigma-Aldrich is committed to providing you powerful and unique screening formats that fully leverage the discovery potential of the trusted and proven TRC shRNA collection. When you partner with Sigma Life Sciences, you gain access to our world class lentiviral production expertise and the formats you require for systematic or pooled RNAi screening on any scale
Genome Triangle
Interested in genome-wide discovery work? LentiElite™, the entire pre-arrayed mouse library, is available in either 96- or 384-well formats. For rapid, whole-genome pooled shRNA screens, LentiPlex® pooled libraries are the answer. Pooled screening presents the entire library in ten vials, and affords expeditious phenotypic, positive selection screens. If you're more interested in focused screening relating to a specific gene family, pathway, process, function or disease relating to your biological question, we offer a host of pre-designed gene family sets and panels to make your screening easier. If you are looking for a quick Human Kinase screening tool, LentiExpress™, a reverse transduction method is available. Do you know exactly the gene you need? Explore YFG to find individual clones for your gene of interest!
From whole genome libraries to individual clones, the MISSION TRC shRNA Lentiviral collection is your path to unlimited discovery.
Wide Array of Product Formats
  • Arrayed Libraries
  • Pooled Libraries
  • Family Sets
  • Reverse Transduction Sets
  • Single Clones
  • Glycerol, DNA and Viral Formats
back to top
Flexible Custom Capabilities
Sigma-Aldrich is the exclusive distributor of the TRC shRNA library clones in DNA or lentiviral format. With world-class lentiviral manufacturing expertise and industry leading custom flexibility, Sigma provides everything you need for even the most unique RNAi project. Additionally, a number of customizable options (including high-throughput projects, custom cloning and large-scale/high titer lentivirus) are available at your fingertips with our new improved ordering interface.
  • Custom Cloning
  • Large-Scale and High Titer Lentiviral Particles
  • Send Your Clones for Lentiviral Manufacturing
  • High-throughput DNA and Lentiviral Manufacturing
  • Custom Gene Family Sets
  • QC Assays and Specification
  • Packaging Materials
  • Other Options
Lab Coat All custom requests will be considered.
Rarely, one size fits all. Small, medium or large — your research needs are your own. So, our technology comes custom-fit with your basic and applied research in mind.