|
For Rapid, Convenient Genome-wide shRNA Screens
MISSION LentiPlex Pooled shRNA Libraries bring the power of the MISSION TRC shRNA collection together with Sigma's lentiviral manufacturing expertise to enable genome-wide RNAi screens on your bench-top. The LentiPlex pooled system provides enhanced delivery and long-term gene silencing in non-dividing and primary cell lines allowing unlimited discovery potential. The convenient format now makes rapid whole-genome RNAi screens accessible to any researcher with minimal reagent, time or capital equipment investment.
- Comprehensive – Trusted TRC content with shRNA targeting 15,000+ human or 15,000+ mouse genes
- Quality – Proprietary methodology ensures uniformity and complete library coverage
- Universal – Simplified whole genome RNAi screening in standard or difficult to expand cell lines
- Accessible – Bench top discovery without robotics or liquid handling system
- Versatile – Compatible with FACS and most downstream fluorescent assays
| Ordering Information |
| Product Description |
Product Number |
| MISSION LentiPlex Human Pooled shRNA Library |
SHPH01 |
| MISSION LentiPlex Mouse Pooled shRNA Library |
SHPM01 |
| Contact your local sales representative for pricing and availability. |
MISSION custom pooled viral shRNA libraries
Pooled shRNA RNAi screening approaches are fast becoming an economical alternative for cell based screens. MISSION RNAi is now offering custom pooled shRNA viral libraries. Just give us your list of genes or pick a gene set and leave the rest to us!
Contact your local Sigma® sales representative for more information or submit an inquiry to MISSIONRNAi@sial.com.
Go directly to: The MISSION LentiPlex System Overview Flow Sorting of Cells Transduced With the MISSION LentiPlex Pooled shRNA Library Product Specifications Contact Us
The MISSION LentiPlex Pooled shRNA Library System Overview The LentiPlex Pooled shRNA Libraries contain the entire TRC collection in 10 matrix tubes. The pooled virus is added to your cell population at a predetermined MOI. Cells are incubated then exposed to selective pressure and selected based on desired phenotype or flow sorting. After selection, cells are then isolated by colony cloning or FACS and the integrated shRNA is identified via PCR, microarray or "Next Generation" sequencing. The identity of potential "hits" can then be used to develop hypotheses regarding the biological role of the corresponding gene(s).
Figure 1. Flow Sorting of Cells Transduced with the MISSION LentiPlex Pooled shRNA Library J774 murine macrophage-like cells were transduced with the pooled mouse shRNA library and analyzed by flow cytometry. Transduced cells were selected in puromycin prior to FACS analysis. Both transduced and untransduced cells were stained with a mixture of FITC- and APC-conjugated primary antibodies. Cells with strong APC and weaker FITC staining were discovered and isolated as "hits" in the screen. These cells are indicated by the arrow in the upper left side of the contour plot (right). The TRC-designed pLKO.1 vector of the LentiPlex pooled constructs enables the isolation of populations using most common fluorescent reporters including FITC and GFP.
Product Specifications The MISSION LentiPlex Human and Mouse Pooled shRNA Libraries are genome-wide lentiviral pools produced using a proprietary process. The libraries each consist of over 75,000 shRNA constructs targeting 15,000+ human or 15,000+ mouse genes. Each library is tested for shRNA representation before product release to ensure robust library coverage. Each library is provided in ready-to-use lentiviral format at titers of at least 5 x 108 TU/ml via p24 assay and is pre-divided into 10 subpools of approximately 8,000 shRNA constructs each. Amplification and sequencing primers are also provided for downstream target identification.
- Pre-aliquoted into 10 subpools of approximately 8,000 shRNA constructs each
- Enough material for six screens at 100X representation
- Titers of at least 5 x 108 TU/ml
- Amplification and sequencing primers provided
Contact Us For questions about ExpressMag or other MISSION lentiviral/RNAi products, please email us at: RNAi@sial.com.