Screening often lies at the beginning of most discovery research. The network of possible molecular interactions leading to the causal phenotype within a given system, pathway or disease is astounding. To better understand this complexity, researchers are moving toward large screening projects to identify candidate genes involved in their biological question of interest. Discovering these novel biological interactions leads to a host of characterization and validation studies, requiring further evaluation. Given the significant amount of follow-up research after a screen, it is essential to begin with the most trusted and comprehensive screening tools available.
Sigma® Life Sciences is committed to providing you powerful and unique screening formats that fully leverage the discovery potential of the trusted and proven TRC shRNA collection. When you partner with Sigma Life Sciences you gain access to our world class lentiviral production expertise and the formats you require for systematic or pooled RNAi screening on any scale. From the LentiElite pre-arrayed whole genome lentiviral shRNA library to individual clones, Sigma offers the complete spectrum of shRNA tools to get you started screening today!
| |
Content |
Format Arrayed – Single shRNA per well Pooled – Multiple shRNAs per well |
Screen Duration +++ = Longer ++ = Moderate + = Quick |
Knockdown Analysis Workload +++ = Advanced ++ = Intermediate + = Novice |
| LentiElite |
Whole Genome, mouse |
Arrayed |
+++ |
+ |
| LentiPlex |
Whole Genome, human or mouse |
Pooled |
+ |
+++* |
| LentiExpress |
Kinome, Human |
Arrayed |
+ |
+ |
| Pre-Designed Panels |
Focused Gene Collections, human or mouse |
Arrayed |
++ |
+ |
Custom-Designed Panels |
Your Gene List, any species |
Arrayed or Pooled |
+ to +++ |
+ to +++ |
| *Note: Identification of shRNA hits/leads within a pooled shRNA screen requires deconvolution using colony cloning, high-throughput sequencing, or FACS analyses. |
Screening Solutions
LentiElite Arrayed Whole Genome Libraries – MISSION LentiElite libraries provide you the ultimate RNAi screening tool. The entire Mouse TRC lentiviral collection pre-arrayed in 96- or 384-well format, 20 µl aliquots.
- Immediate – Start screening today without spending months to years on scale-up and viral production
- Superior Quality – Screen with confidence when you rely on Sigma's world class lentiviral production expertise
- Accessible – Minimize investments in robotics and dedicated personnel for viral manufacturing
LentiPlex Pooled Whole Genome Libraries – MISSION LentiPlex Lentiviral shRNA Pools enable genome-wide RNAi screens at your bench-top with minimal reagent, time or capital equipment investment. The convenient format combines ~80,000 clones into 10 individual tubes for each species.
- Quality – Proprietary methodology ensures uniformity and complete TRC1 library coverage
- Universal – Simplified whole genome RNAi screening in standard or difficult-to-expand cell lines
- Cost Effective – Bench top discovery without robotics, liquid handling system or dedicated freezers
LentiExpress Arrayed Kinome Library – LentiExpress screens consist of pre-arrayed lentiviral particles targeting the human kinome. We offer this premier ready-to-use product format so you can simply add your cells, and proceed directly to loss-of-function screens!
- Simple – No viral particle preparation. Add your cells and go!
- Economical – Single use format
- Save Time – Get your assay results 1 month earlier
Pre-Designed Panels & Sets Our Pre-Designed Panels and Sets are complete collections of genes related to specific functional classes. The format is designed for target discovery and pathway analysis.
- Focused – Specifically tailored to your research questions
- Comprehensive – Thorough bioinformatic investigation across multiple gene databases to generate these lists
- Rapid – Don’t waste time screening through unwanted genes. Dive straight in!
Custom Capabilities Our experience extends beyond the TRC collection because we contract manufacture numerous private collections and alternate libraries from high-throughput projects to large volumes. A number of customizable options (including high-throughput projects, custom cloning and large-scale/high titer lentivirus) are already available to you, and we will continue to consider your requests to bring you more flexibility.
Individual Clones Find Individual Clones. Our comprehensive shRNA product offering consists of over 315,000 pre-cloned shRNA constructs targeting more than 22,000 human and 20,000+ mouse genes.
- More than 125,000 New Clones Available
- More than 4,100 New Genes Targeted
- More than 30,000 Clones Validated!
Selected Publications
Firestein, R et al. CDK8 is a colorectal cancer oncogene that regulates beta-catenin activity. Nature, 455: 547-51 (2008).
Ebert, BL et al. Identification of RPS14 as a 5q-syndrome gene by RNA interference screen. Nature, 451: 335-9 (2008).
Oberdoerffer, S et al. Regulation of CD45 alternative splicing by heterogeneous ribonucleoprotein, hnRNPLL. Science, 321: 686-91 (2008).
Duan, Z, et al. Lentiviral short hairpin RNA screen of genes associated with multidrug resistance identifies PRP-4 as a new regulator of chemoresistance in human ovarian cancer. Mol Cancer Ther., 7: 2377-85 (2008).
Ji, D, et al. A screen of shRNAs targeting tumor suppressor genes to identify factors involved in A549 paclitaxel sensitivity. Oncol. Rep., 18: 1499-505 (2007).
Moffat J, et al., A Lentiviral RNAi Library for Human and Mouse Genes Applied to an Arrayed Viral High-Content Screen. Cell, 124: 1283-1298 (2006).
Contact Us For questions about the library, pricing and quotes or other concerns, please email us at: RNAi@sial.com. MISSION is a registered trademark of Sigma-Aldrich Co. LLC Label License.