shRNA Screening
Genomic DNA Prep & Deconvolution Services
Easily Identify Hits from Pooled shRNA Screens
Need help with deconvolution of pooled shRNA samples after screening is complete? Sigma provides a pooled shRNA deconvolution service that will help you identify TRC clones that are of significance in your screen.
Why Use Sigma as your Deconvolution Partner?
  • Quick turnaround of data (~8 weeks)
  • Consistency in DNA preparation and sequencing
  • Cost effective due to ability to multiplex samples on a single lane 
  • Experience with shRNA pool deconvolution of TRC1, TRC1.5 and TRC2 libraries; including simultaneous sequencing through our proprietary method
  • Data provided in an easy-to-analyze format
Interested? Click the button below and one of our sales reps will help you with your deconvolution needs.
Request Contact
Use the form below to guide the deconvolution discussion with your Sigma® Life Science representative.
Deconvolution Sample Submission Worksheet
Deconvolution Sample Submission Worksheet
(628 Kb PDF)
How it works:
Pooled Screening Approach to Identify Modulators of a Pathway
Get your pooled shRNA from us, conduct your pooled screen at a lower cost and let us help you identify the genes that emerge from your screen. We will deconvolute your pooled shRNA screen with high-throughput sequencing to identify important genes.
Pooled Screening Approach to Identify Modulators of a Pathway
DNA Preparation and Deconvolution Workflow
Once you provide us with cell or tumor samples post screening, we will extract genomic DNA, conduct our proprietary sequencing reaction and provide data on the presence of each TRC clone in each sample. Using our proprietary sequencing technique, we can decode the entire TRC library within a single sequencing reaction. Now we are able to serve your entire pooled shRNA screening workflow.
DNA Preparation and Deconvolution Workflow
Why Use Next-Gen Sequencing Over Other Methods for Pooled Screen Deconvolution?
  • Massive data collection without TA cloning
  • Greater dynamic range
  • Look for enriched or reduced shRNA
  • No hybridization bias, reducing false negatives
  • No cross-hybridization, reducing false positives
  • Precise count of shRNA frequency
  • Detects very low to high abundance shRNA
  • Complete analysis without isolating single cells
  • Analyze multiple pools simultaneously
  • Can be conducted on custom and genome-wide pools
Example of Data Provided from Next-Gen Sequencing
Example of Data Provided from Next-Gen Sequencing