Sigma-Aldrich
Life Science > Functional Genomics & RNAi > shRNA > TRC shRNA Products > MISSION® Lentiviral Controls
Functional Genomics & RNAi
MISSION® Lentiviral Controls
When conducting shRNA experiments, proper controls are a key element of experimental design. Proper use of controls permits accurate interpretation of knockdown results and provides assurance of the specificity of the observed phenotype. Sigma® offers a variety of positive, negative, and transduction controls to meet these needs. All controls are available in both purified plasmid DNA and lentiviral particle format. Controls in DNA format may be used in direct transfection-based experiments or may be used in conjunction with the MISSION® Lentiviral Packaging Mix (SHP001) to create replication incompetent viral particles for use in transduction-based experiments. In addition, our most popular controls are now offered in high titer format "off the shelf!" These controls are provided in lentiviral format with a minimum titer of 109 TU/ml (p24 assay). All other controls, as well as any MISSION® TRC shRNA, can be prepared in high titer format through our custom lentiviral production process.
The MISSION® research and development team, in collaboration with the RNAi Consortium (TRC) scientists, have generated a series of controls to enable successful implementation of your shRNA experiments. The base vector, TRC1.5-pLKO.1-puro1 was developed at the Broad Institute as part of TRC. TRC1.5 is exclusive to Sigma Life Science and contains almost 200,000 clones including more than 49,000 validated clones. It combines all of the content from TRC1 plus an additional 39,212 clones targeting 2,661 new human genes and 2.395 mouse genes. TRC1.5 clones are in the exact same vector backbone as TRC1 clones.
Sigma-Aldrich is both a collaborative member and supplier of TRC1.5 and TRC2. The TRC2 vector is slightly different. The only change between the TRC1.5 vector and the TRC2 vector is the addition of the WPRE (or the Woodchuck Hepatitis Post-Transcriptional Regulatory Element).2 Because there are two vector backbones, we also provide two sets of controls for the best experimental results.
Go directly to:
Recommended Uses, TRC1.5 Controls
TRC1.5 Control Descriptions 
     TRC1.5 Negative Controls
     TRC1.5 Positive Controls for Transduction/Transfection
     TRC1.5 Positive Controls for Knockdown
Recommended Uses, TRC2 Controls
TRC2 Control Descriptions 
     TRC2 Negative Controls 
     TRC2 Positive Controls for Transduction/Transfection
     TRC2 Positive Controls for Knockdown
Recommended Uses, TRC1.5 Controls
MISSION® shRNA Controls Chart Image
back to top
TRC1.5 Control Descriptions
Note: Vector maps for all controls can be found on the shRNA Vector Maps page.
TRC1.5 Negative Controls
Untreated Cells
The first recommendation for a negative control for any good experiment is untreated cells. Untreated cells will provide a reference point for comparing all other samples.
Empty Vector Control (SHC001, SHC001V, SHC001H)
The empty vector, TRC1.5-pLKO.1-puro, is a useful negative control that will not activate the RNAi pathway because it does not contain an shRNA insert. It will allow for observation of cellular effects of the transfection process or the delivery of the lentiviral vector. Cells transfected with the empty vector provide a useful reference point for comparing specific knockdown.
Non-Targeting shRNA Control (SHC002, SHC002V, SHC002H)
This non-targeting shRNA vector is a useful negative control that will activate RISC and the RNAi pathway, but does not target any human or mouse genes. The short-hairpin sequence contains 5 basepair mismatches to any known human or mouse gene. This allows for examination of the effects of shRNA transduction and RNAi activation on gene expression. Cells transfected with the non-target shRNA vector will also provide a useful reference for interpretation of knockdown.
TurboGFP shRNA (SHC004, SHC004V, SHC004H), eGFP shRNA (SHC005, SHC005V), and Luciferase shRNA (SHC007, SHC007V)
Because these shRNAs do not target any known human or mouse genes, they can be used as non-targeting controls in many shRNA experiments.
TRC1.5 Positive Controls for Transduction/Transfection
Sigma now offers a wide variety of controls with fluorescent reporters for the TRC1.5-pLKO.1-puro backbone. Additionally, we offer two fluorescent reporter controls with an alternate promoter driving the fluorophores, Ubiquitin C (UbC). The table and figure below show the excitation and emission wavelengths for the fluorescent proteins used in Sigma’s fluorescent protein controls.
Excitation and Emission Wavelengths for Fluorescent Proteins
Catalog Number Fluorophore Excitation (nm) Emission (nm)
SHC003 / SHC014 TurboGFP 482 502
SHC010 TagCFP 458 480
SHC011 TagYFP 508 524
SHC012 TagRFP 555 584
SHC013 / SHC015 TagFP635 588 635
Excitation and Emisson Spectra for Fluorescent Proteins Image
TurboGFP Control vector (SHC003, SHC003V, SHC003H)
This vector is a useful control for measuring transfection efficiency and optimizing shRNA delivery. The TurboGFP™ Control vector consists of the lentiviral backbone vector, pLKO.1-puro, containing a gene encoding TurboGFP, driven by the CMV promoter. Transfection of this vector provides fast visual confirmation of successful transfection and delivery.
turboGFP-expressionCells expressing TurboGFP 48 hours post-transduction. Lentiviral titer was estimated to be
1.0 x 106 TU/ml using a p24 based ELISA.
TagCFP Control vector (SHC010, SHC010V)
This is a useful control for measuring transduction efficiency and optimizing shRNA delivery. The TagCFP™ Control vector consists of the lentiviral backbone vector, pLKO.1-puro, containing a gene encoding TagCFP, driven by the CMV promoter. This control provides visual confirmation of successful transduction.
CMV-TagCFP™ Image
Cells expressing TagCFP 6 days post-transduction at an MOI of 5.
TagYFP Control vector (SHC011, SHC011V)
This is a useful control for measuring transduction efficiency and optimizing shRNA delivery. The TagYFP™ Control vector consists of the lentiviral backbone vector, pLKO.1-puro, containing a gene encoding TagYFP, driven by the CMV promoter. This control provides visual confirmation of successful transduction.
CMV-TagYFP™ Image
Cells expressing TagYFP 6 days post-transduction at an MOI of 5.
TagRFP Control vector (SHC012, SHC012V)
This is a useful control for measuring transduction efficiency and optimizing shRNA delivery. The TagRFP™ Control vector consists of the lentiviral backbone vector, pLKO.1-puro, containing a gene encoding TagRFP, driven by the CMV promoter. This control provides visual confirmation of successful transduction.
CMV-TagRFP™ Image
Cells expressing TagRFP 6 days post-transduction at an MOI of 5.
TagFP635 Control vector (SHC013, SHC013V)
This is a useful control for measuring transduction efficiency and optimizing shRNA delivery. The TagFP635™ Control vector consists of the lentiviral backbone vector, pLKO.1-puro, containing a gene encoding TagFP635, driven by the CMV promoter. This control provides visual confirmation of successful transduction.
CMV-TagFP635™ Image
Cells expressing TagFP635 6 days post-transduction at an MOI of 5.
UbC-TurboGFP Control vector (SHC014, SHC014V)
This is a useful control for measuring transduction efficiency and optimizing shRNA delivery. The TurboGFP™ Control vector consists of the lentiviral backbone vector, pLKO.1-puro, containing a gene encoding TurboGFP, driven by the UbC promoter. This control provides visual confirmation of successful transduction.
UbC-TurboGFP™ Image
Cells expressing UbC-TurboGFP 6 days post-transduction at an MOI of 5.
UbC-TagFP635 Control vector (SHC015, SHC015V)
This is a useful control for measuring transduction efficiency and optimizing shRNA delivery. The TagFP635™ Control vector consists of the lentiviral backbone vector, pLKO.1-puro, containing a gene encoding TagFP635, driven by the UbC promoter. This control provides visual confirmation of successful transduction.
UbC-TagFP635™ Image
Cells expressing UbC-TagFP635 6 days post-transduction at an MOI of 5.
TRC1.5 Positive Controls for Knockdown
TurboGFP shRNA (SHC004, SHC004V, SHC004H)
The TurboGFP shRNA vector consists of the TRC1.5-pLKO.1–puro vector, containing shRNA that targets TurboGFP, and can be used as a positive control to quickly visualize knockdown. This TurboGFP shRNA control has been experimentally shown to reduce GFP expression by 99.6% in HEK293T cells after 24 hours.
eGFP shRNA (SHC005, SHC005V)
The eGFP shRNA vector consists of the TRC1.5-pLKO.1–puro vector, containing shRNA that targets eGFP (GenBank Accession No. pEGFP U55761), and can be used as a positive control to quickly visualize knockdown.
100 ng of eGFP expression plasmid alone  100 ng eGFP plasmid + 200 ng MISSION eGFP shRNA Control Vector Bright field illumination
Knockdown efficiency of eGFP shRNA in HEK293T cells. Images shown 48 hours post-transfection. A) 100 ng of eGFP expression plasmid alone. B) 100 ng eGFP plasmid + 200 ng MISSION eGFP shRNA Control Vector. C) Bright field illumination.
Luciferase shRNA (SHC007, SHC007V)
The MISSION Luciferase shRNA vector consists of the TRC1.5-pLKO.1-puro vector, containing an shRNA insert that targets luciferase from North American Firefly, Photinus pyralis (GenBank Accession No. M15077). This control can be used as a positive control to quickly confirm knockdown.
Human Positive Control #1, B2M shRNA (SHC008, SHC008V)
B2M is a widely expressed gene, and the protein is found on the surface of most cells. This control will provide clear and measurable knockdown of the human gene target, typically 80-90% in A549 cells, a human epithelial lung carcinoma cell line. In our experiments, we used an MOI of 3, and cells were placed under puromycin selection 1 day post-transduction and grown for 1-2 weeks. The RNA analysis was done using B2M-specific primer probe sets. Western blot data also showed knockdown with this control.
Human Positive Control #2, ARHGDIA shRNA (SHC009, SHC009V)
This control will provide clear and measurable knockdown of the human gene target, typically 80-90% in A549 cells, a human epithelial lung carcinoma cell line. Rho GDP Dissociation Inhibitor (GDI) alpha (ARHGDIA) is a widely expressed gene, and the protein is found intracellularly in most cells.
ARHGDIA ImageA549 cells were transduced with either the empty vector control, SHC001V, or human positive control #2, SHC009V. Cell lysates were then prepared. After determining that mRNA levels had been depleted via hyperbranched PCR, protein levels were probed via Western Blot using anti-ARGHDIA antibody, R3025. Nearly total elimination of protein was observed.
back to top
Recommended Uses, TRC2 Controls
MISSION® TRC2 Controls Chart Image
back to top
TRC2 Control Descriptions
Note: Vector maps for all controls can be found on the shRNA Vector Maps page.
TRC2 Negative Controls
Untreated Cells
The first recommendation for a negative control for any good experiment is untreated cells. Untreated cells will provide a reference point for comparing all other samples.
Empty Vector Control (SHC201, SHC201V)
The empty vector, TRC2-pLKO-puro, is a useful negative control that will not activate the RNAi pathway because it does not contain an shRNA insert. It will allow for observation of cellular effects of the transfection process or the delivery of the lentiviral vector. Cells transfected with the empty vector provide a useful reference point for comparing specific knockdown.
Non-Targeting shRNA Control (SHC202, SHC202V)
This non-targeting shRNA vector is a useful negative control that will activate RISC and the RNAi pathway, but does not target any human or mouse genes. The short-hairpin sequence contains 5 basepair mismatches to any known human or mouse gene. This allows for examination of the effects of shRNA transduction and RNAi activation on gene expression. Cells transfected with the non-target shRNA vector will also provide a useful reference for interpretation of knockdown.
TurboGFP shRNA (SHC204, SHC204V)
Because the shRNA to TurboGFP does not target any known human or mouse genes, it can be used as a non-targeting control in many shRNA experiments.
TRC2 Positive Control for Transduction/Transfection
TurboGFP Control vector (SHC203, SHC203V)
This vector is a useful control for measuring transfection efficiency and optimizing shRNA delivery. The TurboGFP™ Control vector consists of the lentiviral backbone vector, TRC2-pLKO-puro, containing a gene encoding TurboGFP, driven by the CMV promoter. Transfection of this vector provides fast visual confirmation of successful transfection and delivery.
TRC2 Positive Control for Knockdown
TurboGFP shRNA (SHC204, SHC204V)
The TurboGFP shRNA vector consists of the TRC2-pLKO–puro vector, containing shRNA that targets TurboGFP, and can be used as a positive control to quickly visualize knockdown. This TurboGFP shRNA control has been experimentally shown to reduce GFP expression by 99.6% in HEK293T cells after 24 hours.
References
  1. Stewart, S. A. et al. Lentivirus-delivered stable gene silencing by RNAi in primary cells. RNA 2003, 9, 493–501.
  2. Donello, J. E. et al. Woodchuck hepatitis virus contains a tripartite posttranscriptional regulatory element. J. Virol. 1998, 72, 5085-92.
MISSION is a registered trademark of Sigma-Aldrich Co. LLC
TurboGFP, TagCFP, TagYFP, TagRFP, TagFP635 are trademarks of Evrogen.