Sigma-Aldrich
Life Science > Functional Genomics & RNAi > siRNA > MISSION® Positive Control siRNA
siRNA
MISSION® Positive Control siRNA
MISSION® Positive Control siRNA, designed using the Rosetta siRNA Design Algorithm, serve as an ideal complement to any siRNA silencing experiment. All MISSION® Positive Control siRNA have been validated to work*, so you can spend more time silencing your gene of interest and less time on optimizing internal controls.
Benefits of MISSION® Positive Control siRNA:
  • Simplified optimization of siRNA transfection efficiency
  • Increased target specificity, due to siRNA seed region optimization rules
  • Validated gene silencing
  • High quality siRNA produced at ISO 9000:2001 certified sites
  • Freedom to operate for research use
Available MISSION® Positive Control siRNA
Human
Product Gene
GAPDH glyceraldehyde-3-phosphate dehydrogenase
MAPK1 mitogen-activated protein kinase 1
KIF11 kinesin family member 11
Lamin A/C lamin A/C
Cyclophilin B (PPIB) peptidylprolyl isomerase B (cyclophilin B)
TP53 tumor protein p53 (Li-Fraumeni syndrome)
Rat
Product Gene
GAPDH glyceraldehyde-3-phosphate dehydrogenase
MAPK1 mitogen-activated protein kinase 1
Lamin A/C lamin A/C
Cyclophilin B (PPIB) peptidylprolyl isomerase B
TP53 tumor protein p53
*Mission Positive Control siRNA reduce the level of gene target by >75%, 24 hours after transfection with 30nM siRNA (HeLa or 9L/lacZ cells).
Back to siRNA