|
|||||||||||
|
|||||||||||
|
MISSION® Vérité siRNA MISSION® Vérité siRNA utilize the most advanced modification strategy with proven significant reduction in off-target effects while maintaining highly effective specific gene knockdown. Knockdown only the genes you want
It can be difficult to obtain high quality data from RNAi screens because of off-target differential gene expression. These off-target effects are primarily the result of "miRNA-like" seed region binding of the siRNA to unwanted targets. MISSION Vérité siRNA includes a novel RNA base in the seed region of the antisense strand, which significantly reduces overall off-target effects. Looking for significant reduction in seed-region based off-target effects and highly effective specific gene knockdown in your RNAi experiments? Try MISSION Vérité siRNA! What is MISSION Vérité siRNA?
MISSION Vérité siRNA offers the following benefits: Significant reduction in off-target effects
No effect on toxicity or cell viability
Next generation siRNA without the high price
MISSION Vérité siRNA: Effective at standard siRNA concentrations
Efficient target gene knockdown
Works with normal transfection methods and reagents
MISSION Vérité siRNA availability: Custom MISSION Vérité siRNA
Predesigned MISSION Vérité siRNA
Want to find out more about MISSION® Vérité siRNA? Email: Verite@sial.com Contact Us MISSION is a registered trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co. Label License.
|
|||||||||||
|
|
|||||||||||
|