Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Functional Genomics & RNAi > TargeTron > TargeTron® Resources > pACD4K-C Vector Sequence
TargeTron Resources
pACD4K-C Vector Sequence
pACD4K-C Vector Sequence (7,678 bp) Substitute your TargeTron oligo sequences to replace the N’s at the following positions. 
IBS: 1792-1844 (53 bp) EBS2: 2024-2072 (49 bp) EBS1d: 2085-2144 (60 bp) GAATTCCGGATGAGCATTCATCAGGCGGGCAAGAATGTGAATAAAGGCCGGATAAAACTTGTGCTTATTTTTCTTTACGGTCTTTAA
AAAGGCCGTAATATCCAGCTGAACGGTCTGGTTATAGGTACATTGAGCAACTGACTGAAATGCCTCAAAATGTTCTTTACGATGCCA
TTGGGATATATCAACGGTGGTATATCCAGTGATTTTTTTCTCCATTTTAGCTTCCTTAGCTCCTGAAAATCTCGATAACTCAAAAAATA
CGCCCGGTAGTGATCTTATTTCATTATGGTGAAAGTTGGAACCTCTTACGTGCCGATCAACGTCTCATTTTCGCCAAAAGTTGGCCCA
GGGCTTCCCGGTATCAACAGGGACACCAGGATTTATTTATTCTGCGAAGTGATCTTCCGTCACAGGTATTTATTCGGCGCAAAGTGC
GTCGGGTGATGCTGCCAACTTACTGATTTAGTGTATGATGGTGTTTTTGAGGTGCTCCAGTGGCTTCTGTTTCTATCAGCTGTCCCT
CCTGTTCAGCTACTGACGGGGTGGTGCGTAACGGCAAAAGCACCGCCGGACATCAGCGCTAGCGGAGTGTATACTGGCTTACTAT
GTTGGCACTGATGAGGGTGTCAGTGAAGTGCTTCATGTGGCAGGAGAAAAAAGGCTGCACCGGTGCGTCAGCAGAATATGTGAT
ACAGGATATATTCCGCTTCCTCGCTCACTGACTCGCTACGCTCGGTCGTTCGACTGCGGCGAGCGGAAATGGCTTACGAACGGGG
CGGAGATTTCCTGGAAGATGCCAGGAAGATACTTAACAGGGAAGTGAGAGGGCCGCGGCAAAGCCGTTTTTCCATAGGCTCCGC
CCCCCTGACAAGCATCACGAAATCTGACGCTCAAATCAGTGGTGGCGAAACCCGACAGGACTATAAAGATACCAGGCGTTTCCCC
CTGGCGGCTCCCTCGTGCGCTCTCCTGTTCCTGCCTTTCGGTTTACCGGTGTCATTCCGCTGTTATGGCCGCGTTTGTCTCATTCCA
CGCCTGACACTCAGTTCCGGGTAGGCAGTTCGCTCCAAGCTGGACTGTATGCACGAACCCCCCGTTCAGTCCGACCGCTGCGCCT
TATCCGGTAACTATCGTCTTGAGTCCAACCCGGAAAGACATGCAAAAGCACCACTGGCAGCAGCCACTGGTAATTGATTTAGAGG
AGTTAGTCTTGAAGTCATGCGCCGGTTAAGGCTAAACTGAAAGGACAAGTTTTGGTGACTGCGCTCCTCCAAGCCAGTTACCTCGG
TTCAAAGAGTTGGTAGCTCAGAGAACCTTCGAAAAACCGCCCTGCAAGGCGGTTTTTTCGTTTTCAGAGCAAGAGATTACGCGCA
GACCAAAACGATCTCAAGAAGATCATCTTATTAATCAGATAAAATATTTCTAGCTAGATTTCAGTGCAATTTATCTCTTCAAATGTA
GCACCTGAAGTCAGCCCCATACGATATAAGTTGTAATTCTCATGTTTGACAGCTTATCATCGATAAGCTCAAGGAGATGGCGCCC
AACAGTCCCCCGGCCACGGGGCCTGCCACCATACCCACGCCGAAACAAGCGCTCATGAGCCCGAAGTGGCGAGCCCGATCTTC
CCCATCGGTGATGTCGGCGATATAGGCGCCAGCAACCGCACCTGTGGCGCCGGTGATGCCGGCCACGATGCGTCCGGCGTAGA
GGATCGAGATCTCGATCCCGCGAAATTAATACGACTCACTATAGGGGAATTGTGAGCGGATAACAATTCCCCTCTAGNNNNAAG
CTTNNNNNNNNNNNNNNNNNNNNNNNNNNNNNNNNNNNNNNNNNNNTTAAGTCAAGTAGTTTAAGGTACTACTCTGTAAGAT
AACACAGAAAACAGCCAACCTAACCGAAAAGCGAAAGCTGATACGGGAACAGAGCACGGTTGGAAAGCGATGAGTTACCTAAAG
ACAATCGGGTACGACTGAGTCGCAATGTTAATCAGATATAAGGTATAAGTTGTGTTTACNNNNNNNNNNNNNNNNNNNNNNNNN
NNNNNNNNNNNNNNNNNNNNNNNNGAAACCTCTAGTNNNNNNNNNNNNNNNNNNNNNNNNNNNNNNNNNNNNNNNNNNNN
NNNNNTGTACANNNNNTAGGAGAACCTATGGGAACGAAACGAAAGCGATGCCGAGAATCTGAATTTACCAAGACTTAACACTAA
CTGGGGATACCCTAAACAAGAATGCCTAATAGAAAGGAGGAAAAAGGCTATAGCACTAGAGCTTGAAAATCTTGCAAGGGTACGG
AGTACTCGTAGTAGTCTGAGAAGGGTAACGCCCTTTACATGGCAAAGGGGTACAGTTATTGTGTACTAAAATTAAAAATTGATTAG
GGAGGAAAACCTCAAAATGAAACCAACAATGGCAATTTTAGAAAGAATCAGTAAAAATTCACAAGAAAATATAGACGAAGTTTTTA
CAAGACTTTATCGTTATCTTTTACGTCCAGATATTTATTACGTGGCGACGCGTACTAAAACAATTCATCCAGTAAAATATAATATTTT
ATTTTCTCCCAATCAGGCTTGATCCCCAGTAAGTCAAAAAATAGCTCGACATACTGTTCTTCCCCGATATCCTCCCTGATCGACCGG
ACGCAGAAGGCAATGTCATACCACTTGTCCGCCCTGCCGCTTCTCCCAAGATCAATAAAGCCACTTACTTTGCCATCTTTCACAAA
GATGTTGCTGTCTCCCAGGTCGCCGTGGGAAAAGACAAGTTCCTCTTCGGGCTTTTCCGTCTTTAAAAAATCATACAGCTCTCTTG
GGTTAATTGAGGCCTGAGTATAAGGTGACTTATACTTGTAATCTATCTAAACGGGGAACCTCTCTAGTAGACAATCCCGTGCTAAA
TTGTAGGACTGCCCTTTAATAAATACTTCTATATTTAAAGAGGTATTTATGAAAAGCGGAATTTATCAGATTAAAAATACTTTCTCTA
GAGAAAATTTCGTCTGGATTAGTTACTTATCGTGTAAAATCTGATAAATGGAATTGGTTCTACATAAATGCCTAACGACTATCCCTT
TGGGGAGTAGGGTCAAGTGACTCGAAACGATAGACAACTTGCTTTAACAAGTTGGAGATATAGTCTGCTCTGCATGGTGACATGC
AGCTGGATATAATTCCGGGGTAAGATTAACGACCTTATCTGAACATAATGCTTTAAATGGAGTGTCTTCTTCCCAGTTTTCGCAATC
CACATCGGCCAGATCGTTATTCAGTAAGTAATCCAATTCGGCTAAGCGGCTGTCTAAGCTATTCGTATAGGGACAATCCGATATGT
CGATGGAGTGAAAGAGCCTGATGCACTCCGCATACAGCTCGATAATCTTTTCAGGGCTTTGTTCATCTTCATACTCTTCCGAGCAA
AGGACGCCATCGGCCTCACTCATGAGCAGATTGCTCCAGCCATCATGCCGTTCAAAGTGCAGGACCTTTGGAACAGGCAGCTTTC
CTTCCAGCCATAGCATCATGTCCTTTTCCCGTTCCACATCATAGGTGGTCCCTTTATACCGGCTGTCCGTCATTTTTAAATATAGGT
TTTCATTTTCTCCCACCAGCTTATATACCTTAGCAGGAGACATTCCTTCCGTATCTTTTACGCAGCGGTATTTTTCGATCAGTTTTTT
CAATTCCGGTGATATTCTCATTTTAGCCATTTATTATTTCCTTCCTCTTTTCTACAGTATTTAAAGATACCCCAAGAAGCTAATTATAA
CAAGACGAACTCCAATTCACTGTTCCTTGCATTCTAAAACCTTAAATACCAGAAAACAGCTTTTTCAAAGTTGTTTTCAAAGTTGGC
GTATAACATAGTATCGACGGAGCCGATTTTGAAACGCGTTGGGAAATGGCAATGATAGCGAAACAACGTAAAACTCTTGTTGTATG
CTTTCATTGTCATCGTCACGTGATTCATAAACACAAGTGAATGTCGACAGTGAATTTTTACGAACGAACAATAACAGAGCCGTATA
CTCCGAGAGGGGTACGTACGGTTCCCGAAGAGGGTGGTGCAAACCAGTCACAGTAATGTGAACAAGGCGGTACCTCCCTACTTC
ACCATATCATTTTCTGCAGCCCCCTAGAAATAATTTTGTTTAACTTTAAGAAGGAGATATACATATATGGCTAGATCGTCCATTCCG
ACAGCATCGCCAGTCACTATGGCGTGCTGCTAGCGCTATATGCGTTGATGCAATTTCTATGCACTCGTAGTAGTCTGAGAAGGGTA
ACGCCCTTTACATGGCAAAGGGGTACAGTTATTGTGTACTAAAATTAAAAATTGATTAGGGAGGAAAACCTCAAAATGAAACCAAC
AATGGCAATTTTAGAAAGAATCAGTAAAAATTCACAAGAAAATATAGACGAAGTTTTTACAAGACTTTATCGTTATCTTTTACGTCC
AGATATTTATTACGTGGCGTATCAAAATTTATATTCCAATAAAGGAGCTTCCACAAAAGGAATATTAGATGATACAGCGGATGGCT
TTAGTGAAGAAAAAATAAAAAAGATTATTCAATCTTTAAAAGACGGAACTTACTATCCTCAACCTGTACGAAGAATGTATATTGCAA
AAAAGAATTCTAAAAAGATGAGACCTTTAGGAATTCCAACTTTCACAGATAAATTGATCCAAGAAGCTGTGAGAATAATTCTTGAA
TCTATCTATGAACCGGTATTCGAAGATGTGTCTCACGGTTTTAGACCTCAACGAAGCTGTCACACAGCTTTGAAAACAATCAAAAG
AGAGTTTGGCGGCGCAAGATGGTTTGTGGAGGGAGATATAAAAGGCTGCTTCGATAATATAGACCACGTTACACTCATTGGACTC
ATCAATCTTAAAATCAAAGATATGAAAATGAGCCAATTGATTTATAAATTTCTAAAAGCAGGTTATCTGGAAAACTGGCAGTATCAC
AAAACTTACAGCGGAACACCTCAAGGTGGAATTCTATCTCCTCTTTTGGCCAACATCTATCTTCATGAATTGGATAAGTTTGTTTTA
CAACTCAAAATGAAGTTTGACCGAGAAAGTCCAGAAAGAATAACACCTGAATATCGGGAGCTCCACAATGAGATAAAAAGAATTT
CTCACCGTCTCAAGAAGTTGGAGGGTGAAGAAAAAGCTAAAGTTCTTTTAGAATATCAAGAAAAACGTAAAAGATTACCCACACTC
CCCTGTACCTCACAGACAAATAAAGTATTGAAATACGTCCGGTATGCGGACGACTTCATTATCTCTGTTAAAGGAAGCAAAGAGGA
CTGTCAATGGATAAAAGAACAATTAAAACTTTTTATTCATAACAAGCTAAAAATGGAATTGAGTGAAGAAAAAACACTCATCACAC
ATAGCAGTCAACCCGCTCGTTTTCTGGGATATGATATACGAGTAAGGAGATCTGGAACGATAAAACGATCTGGTAAAGTCAAAAA
GAGAACACTCAATGGGAGTGTAGAACTCCTTATTCCTCTTCAAGACAAAATTCGTCAATTTATTTTTGACAAGAAAATAGCTATCCA
AAAGAAAGATAGCTCATGGTTTCCAGTTCACAGGAAATATCTTATTCGTTCAACAGACTTAGAAATCATCACAATTTATAATTCTGA
ACTCCGCGGGATTTGTAATTACTACGGTCTAGCAAGTAATTTTAACCAGCTCAATTATTTTGCTTATCTTATGGAATACAGCTGTCT
AAAAACGATAGCCTCCAAACATAAGGGAACACTTTCAAAAACCATTTCCATGTTTAAAGATGGAAGTGGTTCGTGGGGGATCCCG
TATGAGATAAAGCAAGGTAAGCAGCGCCGTTATTTTGCAAATTTTAGTGAATGTAAATCCCCTTATCAATTTACGGATGAGATAAG
TCAAGCTCCTGTATTGTATGGCTATGCCCGGAATACTCTTGAAAACAGGTTAAAAGCTAAATGTTGTGAATTATGTGGGACGTCTG
ATGAAAATACTTCCTATGAAATTCACCATGTCAATAAGGTCAAAAATCTTAAAGGCAAAGAAAAATGGGAAATGGCAATGATAGCG
AAACAACGTAAAACTCTTGTTGTATGCTTTCATTGTCATCGTCACGTGATTCATAAACACAAGTGAATGTCGAGCACCCGTTCTCGG
AGCACTGTCCGACCGCTTTGGCCGCCGCCCAGTCCTGCTCGCTTCGCTACTTGGAGCCACTATCGACTACGCGATCATGGCGACC
ACACCCGTCCTGTGGATCGCCAAGCTCGCCGATGGTAGTGTGGGGTCTCCCCATGCGAGAGTAGGGAACTGCCAGGCATCAAAT
AAAACGAAAGGCTCAGTCGAAAGACTGGGCCTTTCGTTTTATCTGTTGTTTGTCGGTGAACGCTCTCCTGAGTAGGACAAATCCGC
CGGGAGCGGATTTGAACGTTGCGAAGCAACGGCCCGGAGGGTGGCGGGCAGGACGCCCGCCATAAACTGCCAGGCATCAAATT
AAGCAGAAGGCCATCCTGACGGATGGCCTTTTTGCGTTTCTACAAACTCTTCCTGTCGTCATATCTACAAGCCATCCCCCCACAGA
TACGGTAAACTAGCCTCGTTTTTGCATCAGGAAAGCAGAACGCCATGAGCGGCCTCATTTCTTATTCTGAGTTACAACAGTCCGCA
CCGCTGTCCGGTAGCTCCTTCCGGTGGGCGCGGGGCATGACTATCGTCGCCGCACTTATGACTGTCTTCTTTATCATGCAACTCGT
AGGACAGGTGCCGGCAGCGCCCAACAGTCCCCCGGCCACGGGGCCTGCCACCATACCCACGCCGAAACAAGCGCCCTGCACCA
TTATGTTCCGGATCTGCATCGCAGGATGCTGCTGGCTACCCTGTGGAACACCTACATCTGTATTAACGAAGCGCTAACCGTTTTTA
TCAGGCTCTGGGAGGCAGAATAAATGATCATATCGTCAATTATTACCTCCACGGGGAGAGCCTGAGCAAACTGGCCTCAGGCATT
TGAGAAGCACACGGTCACACTGCTTCCGGTAGTCAATAAACCGGTAAACCAGCAATAGACATAAGCGGCTATTTAACGACCCTGC
CCTGAACCGACGACCGGGTCGAATTTGCTTTCGAATTTCTGCCATTCATCCGCTTATTATCACTTATTCAGGCGTAGCACCAGGCG
TTTAAGGGCACCAATAACTGCCTTAAAAAAATTACGCCCCGCCCTGCCACTCATCGCAGTACTGTTGTAATTCATTAAGCATTCTG
CCGACATGGAAGCCATCACAGACGGCATGATGAACCTGAATCGCCAGCGGCATCAGCACCTTGTCGCCTTGCGTATAATATTTGC
CCATGGTGAAAACGGGGGCGAAGAAGTTGTCCATATTGGCCACGTTTAAATCAAAACTGGTGAAACTCACCCAGGGATTGGCTGA
GACGAAAAACATATTCTCAATAAACCCTTTAGGGAAATAGGCCAGGTTTTCACCGTAACACGCCACATCTTGCGAATATATGTGTA
GAAACTGCCGGAAATCGTCGTGGTATTCACTCCAGAGCGATGAAAACGTTTCAGTTTGCTCATGGAAAACGGTGTAACAAGGGTG
AACACTATCCCATATCACCAGCTCACCGTCTTTCATTGCCATACG