Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Functional Genomics & RNAi > TargeTron > TargeTron Vectors
Functional Genomics & RNAi
TargeTron Vectors
The TargeTron Gene Knockout System (TA0100) allows for the creation of prokaryotic knockouts in as little as three days. TargeTron vectors are now available to increase the capabilities and to enhance the flexibility of the system. The TargeTron Gene Knockout System, in conjunction with the new vectors, can be used to generate multiple knockouts, to create knockouts with removable selection or without using selection, to develop knockouts in organisms lacking a source of T7 RNA polymerase and to generate knockouts in additional organisms such as Staphylococcus aureus.
Multiple Knockouts
TargeTron Vector pACD4K-C-loxP (T2826)
pACD4 TargeTron Non-Selection Vector Set (TV0010)
Knockouts with Removable Selection
TargeTron Vector pACD4K-C-loxP (T2826)
Knockouts without Selection
pACD4 TargeTron Non-selection Vector Set (TV0010)
Source of T7 RNA Polymerase
TargeTron Vector pAR1219 (T2076)
Knockouts in Staphylococcus aureus
TargeTron Vector pNL9164 (T6701)
Knockouts in Clostridium perfringens
TargeTron Vector pJIR750ai (T7701)
TargeTron                                    Vector
TargeTron is a registered trademark of InGex, LLC.
TargeTron® Gene Knockout System