Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
TargeTron
TT Vector pAR1219 data
Data illustrating the disruption of lacZ and tonA in E. coli HB101 using TargeTron® Vector pACD4K-C (included in the TargeTron Gene Knockout System - TA0100) and the T7 helper plasmid pAR1219 (T2076).
Figure 1. Mutation of E. coli HB101 using TargeTron vector pACD4K-C and T7 helper plasmid pAR1219. Targetron vector pACD4K-C was re-targeted to disrupt the lacZ and tonA genes. Mutants were selected on kanamycin plates. Intron isnertions were detected by colony PCR using an intron primer (3p-exon-1, 5'-TACGCAGCGGTATTTTTCGATCAG) and gene specific primers for lacZ and tonA. All colonies screened showed the expected insertion.
Figure 2. White phenotype of HB101 lacZ disruptants created using the pACD4K-C TargeTron plasmid with T7 helper plasmid pAR1219.