Sigma-Aldrich
siRNA Design, Quality, and Delivery
Steven Suchyta
Product Management, Biotechnology Division, Sigma-Aldrich
RNA interference (RNAi) is a natural biological mechanism wherein short inhibitory RNA (siRNA) duplexes induce potent inhibition of gene expression. These siRNA duplexes are produced naturally when an enzyme, Dicer, cleaves long double-stranded RNA (dsRNA) into smaller fragments. The resulting 21-23 nucleotide dsRNA fragments, termed siRNAs, then associate with an RNase-containing complex to form the RNA-induced silencing complex (RISC). The RISC unwinds the duplex and releases the sense strand. The RISC-bound antisense strand then serves as a guide for targeting the activated complex to complementary mRNA sequences. This results in subsequent mRNA cleavage and degradation. In effect, only catalytic amounts of siRNA are required for destruction of mRNA, resulting in the knockdown or silencing of the target gene and diminished protein expression.
RNA (dsRNA) into smaller fragments. The resulting 21-23 nucleotide dsRNA fragments, termed siRNAs, then associate with an RNase-containing complex to form the RNA-induced silencing complex (RISC). The RISC unwinds the duplex and releases the sense strand. The RISC-bound antisense strand then serves as a guide for targeting the activated complex to complementary mRNA sequences. This results in subsequent mRNA cleavage and degradation. In effect, only catalytic amounts of siRNA are required for destruction of mRNA, resulting in the knockdown or silencing of the target gene and diminished protein expression.
The siRNA Experimental Workflow
Conclusions
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Quick Links:
Request LiteratureRequest Literature
Signup with Sigma-AldrichNewsletter Subscription
Join the communityJoin our Life Science
Community
RNA Interface
Click to enlarge 
RNA Interference with Mission siRNA
siRNA Workflow
Click to enlarge 
Sigma siRNA Workflow