Figure 2. ZFN-mediated genome editing takes place in the nucleus when a pair of ZFNs targeting the user’s gene of interest (GOI) is delivered transiently into a cell line, either by transfection or electroporation.