Epidermal Growth Factor (EFG)
Product Description Source Recombinant Host Physical Form Assay Cell Type Affected Cat. No.
Epidermal Growth Factor from murine submaxillary gland - lyophilized powder The biological activity is measured in a proliferation assay using BALB/MK cells.
  • ovary cell
  • chrondrocyte
  • endothelial cell
  • granuloma cell
  • mammary epithelial cell
  • vascular endothelial cell
  • fibroblast
  • prostate cell
  • glial cell
  • HeLa cell
  • myeloma cell
E4127 
Epidermal Growth Factor human Escherichia coli lyophilized powder ≥97%SDS-PAGE
  • endothelial cell
  • HeLa cell
  • ovary cell
  • vascular endothelial cell
  • prostate cell
  • myeloma cell
  • fibroblast
  • mammary epithelial cell
  • glial cell
  • granuloma cell
  • chrondrocyte
E9644
LONG EGF human Escherichia coli powder -
  • granuloma cell
  • mammary epithelial cell
  • ovary cell
  • prostate cell
  • HeLa cell
  • endothelial cell
  • vascular endothelial cell
  • chrondrocyte
  • fibroblast
  • glial cell
  • myeloma cell
E4269
Heparin-Binding EGF-Like Growth Factor human Sf21 cells lyophilized powder The biological activity is measured by its ability to stimulate 3H-thymidine incorporation in the EGF-responsive BALB/3T3 cells.
  • smooth muscle cell
  • Balb/C 3T3
E4643
Amphiregulin
Amphiregulin Human
AR
≥97% (SDS-PAGE), recombinant, expressed in Escherichia coli, lyophilized powder, cell culture tested
Associated gene(s): AREG (374)
ED50/EC50: 10-100 ng/mL
Endotoxin
tested
Store at: -20°C
Betacellulin
Betacellulin Human
BTC
≥97% (SDS-PAGE), recombinant, expressed in Escherichia coli, lyophilized powder, cell culture tested
Associated gene(s): BTC (685)
Endotoxin
tested
Store at: -20°C