| Product Description | Source | Recombinant Host | Physical Form | Assay | Cell Type Affected | Product Number |
| Granulocyte Colony-Stimulating Factor | human | Escherichia coli | lyophilized powder | The biological activity is measured in a cell proliferation assay using a murine myeloblastic cell line, NFS-60. | bone marrow precursor mature granulocyte granulocyte colony-forming cell neutrophil |
G0407 |
| Granulocyte Colony-Stimulating Factor | from mouse | Escherichia coli | lyophilized powder | The biological activity is measured in a cell proliferation assay using a murine myeloblastic cell line, NFS-60. | neutrophil mature granulocyte bone marrow precursor granulocyte colony-forming cell |
G8160 |
| Product Description | Source | Recombinant Host | Physical Form | Assay | Cell Type Affected | Product Number |
| Granulocyte-Macrophage Colony-Stimulating Factor | from rat | Escherichia coli | lyophilized powder | The biological activity is measured by its ability to induce proliferation of the factor-dependent mouse cell line, DA3. | human myeloid progenitor cell | G0792 |
| Granulocyte-Macrophage Colony-Stimulating Factor | from mouse | Escherichia coli | lyophilized powder | The EC 50 activity of mouse GM-CSF is tested in culture using murine FDP-1 cells. | - | G0282 |
| Granulocyte-Macrophage Colony-Stimulating Factor | human | Escherichia coli | lyophilized powder | The proliferative activity of human GM-CSF is tested in culture using human TF-1 cells. | - | G5035 |
| Product Description | Source | Recombinant Host | Physical Form | Assay | Cell Type Affected | Product Number |
| Leukemia Inhibitory Factor | from mouse | Escherichia coli | buffered aqueous solution | The proliferative activity of mouse LIF is measured in culture using the M1 murine cell line. | embryonic carcinoma and stem cell leukemic monocyte hematopoietic cell hepatocyte |
L5158 |
| Leukemia Inhibitory Factor | human | Escherichia coli | buffered aqueous solution (pH 7.4) | The proliferative activity of human LIF is measured in culture using the human leukemic cell line, TF-1. | - | L5283 |
| Product Description | Source | Recombinant Host | Physical Form | Assay | Cell Type Affected | Product Number |
| Macrophage Colony-Stimulating Factor | human | Escherichia coli | lyophilized powder | The proliferative activity is tested in culture using the cytokine-dependent murine monocytic cell line M-NFS-60 cells. | monocyte macrophage |
|
| Macrophage Colony-Stimulating Factor | from mouse | Escherichia coli | lyophilized powder | The proliferative activity is tested by the dose-dependent stimulation of the murine monocytic cell line, M-NFS-60. | - | M9170 |
| Product Description | Source | Recombinant Host | Physical Form | Assay | Cell Type Affected | Product Number |
| Stem Cell Factor | human | Escherichia coli | powder | The proliferative activity is tested in culture using TF-1 cells or by the dose dependent proliferation of human MO-7e cells. | primordial germ cell human bone marrow cell hematopoietic progenitor/stem cell melanoblast |
S7901 |
| Stem Cell Factor | from mouse | Escherichia coli | powder | The biological activity is measured by the dose-dependent stimulation of the proliferation of human TF-1 cells. | human bone marrow cell primordial germ cell melanoblast hematopoietic progenitor/stem cell |
S9915 |
| Product Description | Source | Recombinant Host | Physical Form | Assay | Cell Type Affected | Product Number |
| Thrombopoietin | human | Escherichia coli | lyophilized powder | - | - | T1568 |
| Thrombopoietin | from mouse | NSO cells | lyophilized powder | The biological activity was measured in a cell proliferation assay using MO7e cells. | - | T4184 |
| Product Description | Source | Recombinant Host | Physical Form | Assay | Cell Type Affected | Product Number |
| Erythropoietin | from rat | insect cells | - | The biological acitivity is measured by its ability to stimulate cell proliferation using TF-1 cell line. | - | E8905 |
| Erythropoietin | human | CHO cells | lyophilized powder | The biological activity is assayed in tissue culture using mouse fetal liver cells. | - | E5627 |
| Erythropoietin | from mouse | mouse NSO cells | lyophilized powder | The biological acitivity is measured by its ability to stimulate cell proliferation using TF-1 cell line. | - | E9530 |
| Product Description | Source | Recombinant Host | Physical Form | Assay | Cell Type Affected | Product Number |
| Oncostatin M | human | Escherichia coli | lyophilized powder | The proliferative activity of human oncostatin M is tested in culture by using a human erythroleukemic cell line, TF-1. | - | O9635 |
| Oncostatin M | from mouse | Escherichia coli | lyophilized powder | The biological activity of recombinant mouse OSM was measured by its ability to stimulate 3 H-thymidine incorporation in quiescent NIH/3T3 cells. | - | O1637 |