Sigma-Aldrich
Life Science > Learning Center > BioFiles > BioFiles v4 n5 > Insulin-Like Growth Factors
Biofiles Issue 4-5
Insulin-Like Growth Factors
LONG®R3IGF-I: An Insulin Alternative Exclusively for Cell Culture Applications
Download article
LONG®R3IGF-I is a recombinant analog of human insulin-like growth factor-I (IGF-I) that has been specifically engineered for the enhancement of cell culture performance. LONG®R3IGF-I is more biologically potent in vitro than either insulin or native IGF-I and has been shown to significantly increase recombinant protein production. It is ideal for both research and large-scale culture systems utilizing serum-free or low-level serum applications.
LONG®R3IGF-I is specifically designed and manufactured exclusively for the research and industrial cell culture market for distribution by SAFC Biosciences.
LONG®R3IGF-I: Better science for better cell culture
Recombinant human insulin and other growth factors are essential for long-term growth and proliferation of cell lines. Although insulin is used as a growth factor in cell culture, its primary use is as a therapeutic drug for the treatment of diabetes. This has led to supply and availability issues for research and industrial cell culture users. In contrast, LONG®R3IGF-I is a dedicated raw material manufactured exclusively for cell culture applications providing a consistent, compliant and reliable alternative to recombinant insulin. It is not subject to market fluctuations and shortages as is recombinant insulin.
Proven science to maximize cell culture performance
When LONG®R3IGF-I is supplemented in serum-free medium it promotes cell proliferation, increased cell survival, increased productivity through greater proliferation and anti-apoptotic signaling. LONG®R3IGF-I provides equivalent or better performance to recombinant insulin depending on the cell line and clone. LONG®R3IGF-I has been used with numerous cell types including CHO, BHK, HEK 293, Vero, PER.C6®, MDCK and fibroblasts (graphs 1-4). All cell types that have a growth response to insulin in cell culture have the potential to respond to LONG®R3IGF-I.
LONG®R3IGF-I is an analogue of human-like growth factor I (IGF-I) specifically engineered for use in research and industrial cell culture. Insulin, like IGF-I and their receptors (the insulin receptor (IR) and the type-I IGF receptor IGF-IR) have similar amino acid sequence and protein structure. As a consequence, insulin and IGF-I are able to bind to each other's receptor with relatively low affinity. It is widely accepted that in CHO cells the effects of insulin are mediated by the IGF-IR, due to the fact there are relatively few IR present on the CHO cells and that insulin must be present at a high, non-physiological concentration typically 1 -10 mg/L, to be effective. A more effective growth factor is one which targets and activates the IGF-IR directly, such as IGF-I or the analogue LONG®R3IGF-I. LONG®R3IGF-I has a distinct biological advantage over native IGF-I due to its low affinity for IGF Binding Proteins (IGFBPs). All mammalian cells secrete IGFBPs, which bind to and inhibit native IGF-I. The substitution of an arginine for glutamine acid at position three in LONG®R3IGF-I, in conjunction with the 13 amino acid N-terminal extension peptide, results in > 1000-fold reduced affinity for IGFBPs enhancing bioavailability and effectiveness in comparison to native IGF-I.
Increase cell density, maintain higher viability and extend culture duration
Under bioreactor conditions, stress induced apoptosis is the major cause of loss of cell viability. Activation of the IGF-IR results in the stimulation of a number of signal transduction cascades that have been identified as important for cell survival and proliferation. LONG®R3IGF-I not only results in greater activation of IGF-IR over insulin, but also results in greater activation of key anti-apoptotic and proliferative signaling molecules: Akt and MAPK. Increased activation of these signaling molecules prevents the loss of viability caused by different culture conditions.
Prolonged cell culture activity through greater stability
Under normal cell culture conditions, LONG®R3IGF-I is more stable than insulin, persisting up to two times longer. Insulin is degraded in cell culture by enzymes (insulinases) secreted by cells into culture media. In addition, insulin is more rapidly internalized and degraded compared with IGF-I and LONG®R3IGF-I. The extended cell culture stability of LONG®R3IGF-I results in prolonged activity and associated benefits to cell culture. (Graph 5).
Regulatory compliant cGMP and animal free
LONG®R3IGF-I is manufactured under cGMP compliance with the International Conference on Harmonization (ICH) Q7A guidelines. The manufacturing plant is regularly audited by major European, US and Japanese biopharmaceutical companies. LONG®R3IGF-I is produced in a proprietary E. coli fermentation process that is completely animal free (AF). No animal-derived materials are used during the manufacture or storage of LONG®R3IGF-I. A certificate of origin is available upon request. Manufactured by Novozymes Biopharma, LONG®R3IGF-I has been supplied to the research and industrial cell culture market for over 15 years. In addition, LONG®R3IGF-I is used in the manufacture of several FDA (US), EMEA (Europe) and MHLW (Japan) approved marketed products, with many more in late-phase clinical trials.
Product Description Source Recombinant Host Physical Form Assay Cell Type Affected Product Number
LONGR3 IGF-I human Escherichia coli Lyphilized Powder ≥95% SDS-PAGE - 85580C -
Test Specification
Appearance Lyophilized white/creamy crystalline powder or a clear liquid
Endotoxin < 0.10 EU/μ protein
Bioburden Total viable aerobic count ≤100 cfu/mL
Biological Activity ED50 <10 ng/mL (Bioassay assessing the stimulation of protein synthesis in L6 myoblasts)
Concentration 0.9-1.1 mg/mL by reverse-phase HPLC
Identity Confirmed by N-terminal sequence analysis and reverse-phase HPLC (18 residues >95% single sequence)
Purity A single band ≥95% as determined by SDS-PAGE
ORDERING
SERVICE & SUPPORT
CORPORATE
    Follow us on Twitter!  Join us on Facebook!  Join us on LinkedIn!  Watch our videos on YouTube!
Copyrights © 2010 Sigma-Aldrich Co. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.