| Product Description | Source | Recombinant Host | Physical Form | Assay | Cell Type Affected | Product Number |
| Interleukin-1α | from mouse | Escherichia coli | lyophilized powder | The biological activity is measured in a cel proliferation assay using the murine helper T cell line, D10.G4.1. | neutrophil LAK cell megakaryocyte NK cell granulocyte macrophage monocyte murine B and T cell |
|
| Interleukin-1α | from rat | Escherichia coli | lyophilized powder | The biological activity of recombinant rat IL-1α was measured by its ability to stimulate cell proliferation using a mouse helper T cell line, D10.G4.1. | lymphocyte T cell |
I3901 |
| Interleukin-1α | - | Escherichia coli | lyophilized powder | The proliferative activity of human IL-1α is tested in culture using the murine cell line D10S. | T cell lymphocyte |
I2778 |
| Interleukin-1β | from mouse | Escherichia coli | lyophilized powder | The biological activity is measured by the dose-dependent stimulation of murine D10S cells. | macrophage monocyte neutrophil murine B and T cell megakaryocyte granulocyte LAK cell NK cell |
I5271 |
| Interleukin-1β | human | Escherichia coli | lyophilized powder | The biological activity is measured by the dose-dependent stimulation of MTS cleavage by murine D10S cells. | lymphocyte T cell |
I9401 |
| Interleukin-1β | - | Escherichia coli | lyophilized powder | The biological activity is measured in a cell proliferation assay using murine D10S cells. | lymphocyte | I2393 |
| Interleukin-2 | from mouse | Escherichia coli | lyophilized powder | Activity was determined by dose-dependent stimulation of murine CTLL-2 cells. ED50 : ≤ 0.2 ng/ml (corresponds to a specific activity of 5 × 106 units/mg) | murine T cell NK cell |
I0523 |
| Interleukin-2 | - | Escherichia coli | lyophilized powder | The biological activity is measured by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells. | macrophage B cell oligodendrocyte NK cell monocyte LAK cell |
I2644 |
| Interleukin-3 | human | Escherichia coli | lyophilized powder | The biological activity is measured by the dose-dependent stimulation of the proliferation of human TF-1 cells. | macrophage T cell megakaryocyte neutrophil mast cell |
I1646 |
| Interleukin-3 | human | Escherichia coli | lyophilized powder | ≥ 95% SDS-PAGE | neutrophil macrophage megakaryocyte T cell mast cell B cell |
I7389 |
| Interleukin-3 | from mouse | Escherichia coli | lyophilized powder | The biological activity is measured by the dose-dependent stimulation of the proliferation of mouse M-NFS-60 cells. | macrophage T cell B cell mast cell megakaryocyte neutrophil |
I4144 |
| Interleukin-4 | human | Escherichia coli | lyophilized powder | The biological activity is measured by the dose-dependent stimulation of the proliferation of human TF-1 cells. | T cell macrophage mast cell B cell myeloid progenitor cell |
I4269 |
| Interleukin-4 | from mouse | Escherichia coli | lyophilized powder | The proliferative activity is tested in culture using mouse HT-2 cells. | B cell macrophage T cell mast cell |
I1020 |
| Interleukin-5 | from mouse | Sf21 cells | lyophilized powder | - | - | I1145 |
| Interleukin-5 | human | Sf21 cells | lyophilized powder | - | - | I5273 |
| Interleukin-6 | human | - | solution | The bioactivity of IL-6 is determined in a bioassay system of T-1165 cell proliferation compared to NIBSC Reference preparation code 88/514. | fibroblast B cell myeloid progenitor cell hepatocyte T cell |
I3268 |
| Interleukin-6 | human | Escherichia coli | lyophilized powder | - | fibroblast hepatocyte T cell myeloid progenitor cell B cell |
I1395 |
| Interleukin-6 | from mouse | Escherichia coli | lyophilized powder | The biological activity of recombinant mouse IL-6 was measured in a cell proliferation assay using the mouse 7TD1 cells. | myeloid progenitor cell hepatocyte T cell fibroblast B cell |
I9646 |
| Interleukin-6 | - | Escherichia coli | buffered aqueous solution | The biological activity is measured in a cell proliferation assay using IL-6 dependent mouse plasmacytoma cell line, T1165.85.2.1. | B cell | I0406 |
| Interleukin-7 | human | Escherichia coli | lyophilized powder | - | - | I5896 |
| Interleukin-7 | from mouse | Escherichia coli | lyophilized powder | - | - | I4892 |
| Interleukin-9 | human | Sf21 cells | lyophilized powder | The proliferative activity of recombinant human IL-9 is tested in culture using M07e cells. | - | I3394 |
| Interleukin-10 | from mouse | Escherichia coli | buffered aqueous solution | The biological activity of IL-10 is determined in a cell proliferation assay using MC/9 cells. | - | I3019 |
| Interleukin-11 | human | Sf21 cells | lyophilized powder | The biological activity is measured in a cell proliferation assay using T11, a subline of the IL-6-dependent murine plasmacytoma cell line T1165.85.2.1 that has been adapted to gtr in IL-11. | - | I2406 |
| Interleukin-12 | human | Sf21 cells | lyophilized powder | - | - | I2276 |
| Interleukin-12 | from mouse | Sf21 cells | lyophilized powder | - | - | I8523 |
| Interleukin-13 | human | Escherichia coli | lyophilized powder | - | - | I1771 |
| Interleukin-13 | from mouse | Escherichia coli | lyophilized powder | - | - | I1896 |
| Interleukin-15 | human | Escherichia coli | lyophilized powder | The bioactivity of IL-15 is tested with M07e cells. | - | I8648 |