| Bone Morphogenetic Protein 2 |
human |
Escherichia coli |
lyophilized powder |
The biological activity is measured by its ability to induce alkaline phosphatase production by ATDC5 chondrogenic cells. |
- |
B3555 |
| Bone Morphogenetic Protein 4 |
human |
mouse NSO cells |
lyophilized powder |
The biological activity is measured by its ability to induce alkaline phosphatase production by C2C12 myoblasts. |
- |
B2680 |
| Bone Morphogenetic Protein 5 |
human |
mouse NSO cells |
lyophilized powder |
The biological activity is measured by its ability to induce alkaline phosphatase production in ATDC5 chondrogenic cells. |
- |
B9803 |
| Bone Morphogenetic Protein 6 |
human |
mouse NSO cells |
lyophilized powder |
The biological activity is measured by its ability to induce alkaline phosphatase production by MC3T3-E1 cells. |
- |
B2805 |
| Bone Morphogenetic Protein 7 |
human |
CHO cells |
lyophilized powder |
The biological activity is measured by its ability to induce alkaline phosphatase production using mouse ATDC-5 chondrogenic cells. |
- |
B1434 |