| Product Description | Source | Recombinant Host | Physical Form | Assay | Cell Type Affected | Product Number |
| Placenta Growth Factor | human | Escherichia coli | powder | The biological activity is measured by the ability to bind rhFlt-1/Fc in a functional ELISA. Immobilized rhFlt-1/Fc at 2 µg/mL (100 µL/well) can bind rhP/GF with a linear range at 0.1 - 5 ng/mL. | endothelial cell progenitor vascular endothelial cell monocyte |
P1588 |
| Product Description | Source | Recombinant Host | Physical Form | Assay | Cell Type Affected | Product Number |
| Placenta Growth Factor-2 | from mouse | Sf21 cells | lyophilized powder | The biological activity is measured by its ability to bind to immobilized Flt1/Fc chimera in ELISA. | vascular endothelial cell endothelial cell progenitor monocyte |
P5739 |
| Product Description | Source | Recombinant Host | Physical Form | Assay | Cell Type Affected | Product Number |
| Vascular Endothelial Growth Factor | from mouse | Escherichia coli | powder | The biological activity is measured by the ability to stimulate 3H-thymidine incorporation in human umbilical vein endothelial cells (HUVEC) cells and bovine aortic endothelial cells. | glial cell vascular endothelial cell mammary epithelial cell |
V4512 |
| Vascular Endothelial Growth Factor | human | Escherichia coli | powder | - | myeloma cell ovary cell prostate cell chrondrocyte |
V7259 |
| Vascular Endothelial Growth Factor 120 | from mouse | Escherichia coli | lyophilized powder | The biological activity is measured by its ability to stimulate 3H-thymidine incorporation in human umbilical vein endothelial cells. | - | V3513 |
| Vascular Endothelial Growth Factor 121 | human | Escherichia coli | lyophilized powder | The biological activity is measured by its ability to stimulate 3H-thymidine incorporation in human umbilical vein endothelial cells. | - | V3388 |
| Vascular Endothelial Growth Factor 164 | from rat | mouse NSO cells | lyophilized powder | The biological activity is measured by its ability to stimulate 3H-thymidine incorporation in human umbilical vein endothelial cells. | - | V3638 |
| Product Description | Source | Recombinant Host | Physical Form | Assay | Cell Type Affected | Product Number |
| Vascular Endothelial Growth Factor D | human | Sf21 cells | powder | The biological activity of VEGF-D is measured by its ability to bind to recombinant human FLT-4/Fc in a ELISA assay. | granuloma cell endothelial cell HeLa cell |
V6012 |