Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Biowire Header
Biowire Fall 2010 — A Look into the Future of Gene Editing
Download Biowire Fall 2010 (16 Mb PDF)
Table of Contents
Phosphatase Inhibitor Cocktail
A key additive in protein phosphorylation research
Introduction
Protein Phosphatases
Phosphatase Inhibitor Cocktails
Experimental results
Inhibition of Protein Phosphatase 1a-like activity
Inhibition of Alkaline Phosphatase (AP) like activity
Introduction
Phosphorylation is a reversible protein post-translational modification. Protein kinases catalyze the transfer of the negatively charged ?-phosphate group from ATP to a hydroxyl side chain of serine, threonine or tyrosine residues of the protein, changing the protein's conformation and activity. The reverse reaction, phosphate removal from the protein, or dephosphorylation, is performed by protein phosphatases. Three to four percent of mammalian proteins are kinases and phosphatases,1,2 some being specific for a few target proteins while others act broadly on many proteins.
Protein phosphorylation is a key regulatory mechanism that controls protein activity, interactions, localization and degradation. It controls processes of signal transduction, cell cycle, apoptosis, metabolism and others. About one-third of the proteins present in a typical mammalian cell are phosphorylated, many of them on multiple sites. Serine phosphorylation accounts for 86% of the phosphoproteome, while threonine and tyrosine phosphorylation are 12% and 2% respectively. Numerous human diseases are associated with abnormal phosphorylation of cellular proteins.3-5
There are many advantages for a cell to utilize phosphorylation/ dephosphorylation of a protein as a control mechanism. It is rapid, does not require the synthesis of new proteins or degradation of proteins, and is a reversible reaction.
When studying cellular processes involving protein phosphorylation such as signaling cascades and protein-protein interactions, or analysis of phosphorylated sites on a protein and phosphoprotein purification, it is essential to inhibit cellular protein phosphatases by phosphatase inhibitors. This allows freezing of the phosphorylation state of the target protein/s at an elected time point. The phosphorylation database www.phosphosite.org reports 80,000 phospho-sites. The detection of most of them would not have been possible without the use of phosphatase inhibitors.
back to top
Protein Phosphatases
Protein phosphatases are divided to subcategories based on their substrate specificity (see Table 1).
  1. Alkaline phosphatase6-8 – A family of non-specific phosphatases, responsible for removing phosphate groups from many types of molecules, including proteins, nucleotides and alkaloids. Mammalian alkaline phosphatase isoenzymes are inhibited by homoarginine and levamisole analogs. However, the intestinal and placental isoenzymes are not inhibited by levamisole but are inhibited by imidazole.
  2. Protein Serine/Threonine phosphatase9,10 – This class of phosphoprotein phosphatases accounts for the majority of Ser/Thr phosphatase activity in vivo. It includes two major subclasses, PP1 and PP2. The latter is further subdivided based on metal-ion requirement: PP2A that does not require metal ion; PP2B which is calcium stimulated; and PP2C that is Mg2+ dependent. Ser/Thr phosphatases are inhibited by many small molecules from marine sponges, soil streptomyces and others. The best known are okadaic acid, calyculin A, microcystin-LR, tautomycin, fostriecin and cantharidin. The activities of these compounds have different specificities towards various Ser/Thr phosphatases.11
  3. Protein Tyrosine phosphatase12 – A group of enzymes that remove phosphate groups from phosphorylated tyrosine residues on proteins, using a cysteinyl-phosphate enzyme intermediate. These enzymes are key regulatory components in signal transduction pathways. Tyrosine phosphatases are inhibited by orthovanadate and related compounds and by sodium fluoride.
  4. Dual specificity (Tyr and Ser/Thr) phosphatases3 – Dual-Specificity phosphatases are a subclass of protein tyrosine phosphatases that are also able to dephosphorylate both serine and threonine residues. They are involved in the regulation of key cell signaling pathways. Specific examples of these inhibitors can be found in reference.13
Table 1. Types of phosphatases present in animal tissues and cell cultures
Biowire Fall 2010 Phosphatase Inhibitor Cocktail Table 1
"When studying cellular processes involving protein phosphorylation such as signaling cascades and protein-protein interactions, or analysis of phosphorylated sites on a protein and phosphoprotein purification, it is essential to inhibit cellular protein phosphatases by phosphatase inhibitors."
back to top
Phosphatase Inhibitor Cocktails
The use of phosphatase inhibitors is critical for all types of phosphorylation studies. Sigma® Life Science provides a line of phosphatase inhibitor cocktails that cover the widest range of phosphatases to ensure protection of proteins from dephosphorylation. The line contains 2 cocktails: Phosphatase Inhibitor Cocktail 2 (Product No. P5726) and Phosphatase Inhibitor Cocktail 3 (Product No. P0044). Phosphatase Inhibitor Cocktail 3, Sigma's new phosphatase inhibitor cocktail, is a mixture of phosphatase inhibitors directed towards Ser/Thr phosphatases. Phosphatase Inhibitor Cocktail 3 replaces Phosphatase Inhibitor Cocktail 1 (Product No. P2850). The difference between the two cocktails is that the new Phosphatase Inhibitor Cocktail 3 contains Calyculin A instead of microcystin-LR in Phosphatase Inhibitor Cocktail 1. Table 2 provides the list of inhibitors in each cocktail.
Table 2. Phosphatase Inhibitor Cocktail components
Biowire Fall 2010 Phosphatase Inhibitor Cocktail Table 2
back to top
Experimental results
The following experimental data compares the inhibitory effect of the new Phosphatase Inhibitor Cocktail 3 and Phosphatase Inhibitor Cocktail 1 (Product No. P2850) which it replaces. The data shows that the activity of the two cocktails is equivalent.
It is important to mention that in order to inhibit a wide range of phosphatases and to effectively reduce total phosphatase activity, it is recommended to use both of Sigma’s phosphatase inhibitor cocktails, Phosphatase Inhibitor Cocktail 2 (Product No. P5726) and Phosphatase Inhibitor Cocktail 3 (Product No. P0044).
back to top
Inhibition of Protein Phosphatase 1a-like activity
Phosphatase Inhibitor Cocktail 3 (Product No. P0044) is an effective inhibitor of Protein phosphatase 1a-like activity in cells and tissues. The following experiments demonstrate its inhibition efficiency and compare it to that of the former Phosphatase Inhibitor Cocktail 1 (Product No. P2850).
Biowire Fall 2010 Phosphatase Inhibitor Cocktail Figure 1
Figure 1. Inhibition of PP1a-like activity in cell and tissue extracts
Endogenous PP1a-like activity in various cell and tissue extracts was measured using the radioactive substrate 32P-Ser phosphorylase A at pH 7.5, 30°C.20 Phosphatase Inhibitor Cocktail 1 (Product No. P2850) or Phosphatase Inhibitor Cocktail 3 (Product No. P0044) were added to the extracts at a final concentration of 1% at 30°C, 5 minutes prior to assaying PP1a-like activity. The activities shown in Figure 1 are relative and not absolute. The smaller the activity remaining, the greater the degree of inhibition by the cocktail that was placed in the reaction.
* The value observed for these samples is too low to be seen in the diagram and is very close to zero.
Biowire Fall 2010 Phosphatase Inhibitor Cocktail Figure 2
Figure 2. Dose dependent inhibition of PP1a-like activity in human placenta
Human placenta shows 10-20 folds more PP1a-like activity per mg protein than other tissue extracts. Therefore, it was chosen to demonstrate the inhibition efficiency of Phosphatase Inhibitor Cocktails.
Endogenous PP1a-like activity in human placenta extract was measured using the radioactive substrate 32P-Ser phosphorylase A at pH 7.5, 30°C.20 Different amounts of Phosphatase Inhibitor Cocktail 1 (Product No. P2850) or Phosphatase Inhibitor Cocktail 3 (Product No. P0044) were added to the extract at 30°C, 5 minutes prior to assaying PP1a-like activity.
back to top
Inhibition of Alkaline Phosphatase (AP) like activity
As mentioned above, different alkaline phosphatase isoenzymes are inhibited by different inhibitors. Phosphatase Inhibitor Cocktail 3 (Product No. P0044) strongly inhibits the L-isoforms of alkaline phosphatase present in extracts from bovine liver, and shows a synergistic effect when applied with Phosphatase Inhibitor Cocktail 2 (Product No. P5726) (see Figure 3A).
Phosphatase Inhibitor Cocktail 3 (Product No. P0044) has little effect on the inhibition of the P-isoforms of alkaline phosphatase activity, which are abundant in human placental extracts, and thus inhibition is achieved by the use of Phosphatase Inhibitor Cocktail 2 (Product No. P5726) (see Figure 3B).
Biowire Fall 2010 Phosphatase Inhibitor Cocktail Figure 3
Figures 3A & B. Dose dependent Inhibition of Alkaline Phosphatase (AP) like activity in bovine liver extract
Endogenous AP-like activity in bovine liver extract was measured in a colorimetric assay using pNPP as a substrate at pH 10.4, 37°C. Inhibition was performed by incubating the extract with Phosphatase Inhibitor Cocktail 3 (Product No. P0044) alone or in tandem with Phosphatase Inhibitor Cocktail 2 (Product No. P5726), for 3 minutes at 37°C prior to assaying AP-like activity.
back to top
References
  1. Johnson, L.N., Biochemical Society Transactions, 37, 627-641 (2009).
  2. Moorhead, G.B.G., et al., Biochemistry Journal., 417, 401-409 (2009).
  3. Shchemelinin, I., et al., Folia Biologica (Praha), 52, 81-101 (2006).
  4. O lsen, J.V., et al., Cell, 127, 635-648 (2006).
  5. Graves, J.D., and Krebs, E.G., Pharmacology and Therapeutics, 82, 111-121 (1999).
  6. Moss, D.W., Clinical Chemistry, 38, 2486-2492 (1992).
  7. O nsgard-Meyer, M., et al., Proceedings of the Society for Experimental Biology and Medicine, 213, 193-195 (1996).
  8. Crofton, P.M., Critical Reviews in Clinical Laboratory Sciences, 16, 161-194 (1982).
  9. Barford, D., Trends in Biochemical Sciences, 21, 407-412 (1996).
  10. Wera, S., and Hemmings, B.S., Biochemistry Journal, 311, 17-29 (1995).
  11. Swingle, M., et al., Methods in Molecular Biology, 365, 23-38 (2007).
  12. Wang, W-Q., et al., Current Topics in Medicinal Chemistry, 3, 739-748 (2003).
  13. Pestell, K.E., et al., Oncogene, 19, 6607-6612 (2000).
  14. Van Belle, H., et al., Clinical Chemistry, 23, 454-459 (1977).
  15. Chakrabartty, A., and Stinson, R.A., Biochimica et Biophysica Acta, 839, 174-180 (1983).
  16. Brunel, C., and Cathala, G., Biochimica et Biophysica Acta, 268, 415-421 (1972).
  17. Takai, A., and Mieskes, G., Biochemistry Journal, 275, 233-239 (1991).
  18. Lum, H., et al., American Journal of Physiology - Lung Cellular and Molecular Physiology, 281, L546-L555 (2001).
  19. Posner, B.I., et al., Journal of Biological Chemistry, 269, 4596-604 (1994).
  20. Cohen, P., et al., Methods in Enzymology., 159, 390-408 (1988).
back to Biowire Fall 2010
back to top