Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Metabolomics > BioUltra > Antioxidants
BioUltra Reagents
Antioxidants
Antioxidants, also known as protecting agents, are substances which are added to solutions used in biology, biochemistry or microbiology to prevent chemical changes caused by exposure to oxygen. Cultures of anaerobic bacteria may require the presence of substances buffering the redox potential at negative, reducing values. Breaking cells, especially with ultrasonic waves, introduces oxygen into the cell extract, inducing oxidation of cellular compounds such as lipids or the sulfhydryl groups in proteins. As a result, enzymes possessing functional sulfhydryl groups become inactivated. Such negative effects can be minimized by the addition of protecting agents, such as ascorbate, dithiothreitol, mercaptoethanol or cysteine. These chemicals are usually used in concentrations of 10-3 to 10-2 M. Their redox potential lies in the range of 0 to -350 mV at pH 7. Sulfhydryl agents maintain the free sulfhydryl groups of proteins in their reduced state and also reduce previously formed disulfide bridges. Additionally, these agents often display chelating properties, masking heavy metal ions in solution. Good protecting agents should be stable in air and under physiological conditions, and should not be oxidized during experimentation.
View our Antioxidants here.
Redox Properties of Antioxidants
Antioxidant Redox System E0' Ref.
Ascorbic acid Dehydroascorbic acid/Asorbic acid 58 mV [1]
Cysteine Cystine/Cysteine -340 mV [1]
Dithioerythritol C4H8O2S2/C4H10 O2S2 -330 mV [2]
Dithionite SO32-/S2O42- -527 mV [1]
Dithiothreitol C4H8O2S2/C4H10O2 S2 -330 mV [2]
Pyrosulfite   * [3]
E0': Midpoint potential at pH 7.
*: Pyrosulfite in aqueous solution, gives a mixture of pyrosulfite, bisulfite, sulfite, sulfurous acid, and SO2. The proportions depend on temperature, concentration, and pH.
[1] CRC Handbook of Biochemistry and Molecular Biology, 3rd ed., Physical Chemical Data, vol. I, p. 122-130, G.D. Fasman, ed. CRC Press, Cleveland, Ohio 1976
[2] W.W Cleland, Biochemistry 3, 480 (1964)
[3] W.D. Loomis, Methods Enzymol. 31, 528 (1974)
Back to BioUltra home
Back to Bioscience home