Enzyme Explorer
Novozymes
Continuing a commitment to life science and high-technology, Sigma has partnered with Novozymes™ to bring our customers the benefits of the Novozyme line. The world's largest manufacturer of high-quality industrial enzyme products, Novozymes is a 2001 recipient of the Presidential Green Chemistry Challenge Award for eco-friendly products. This partnership makes the Novo product line easily available to researchers through the Sigma-Aldrich Enzyme Explorer.
Novozymes            logo Take advantange of quality brand name enzymes including: Everlase™, Lipolase™, Pectinex™, Glucanex™ and more — all just a click away! No where else can you save money and time, while finding efficient and environmentally-friendly solutions.
Click on the column headings to sort products. To re-alphabetize, hit the page refresh button in your web browser.
Novozyme
Brand Name
Product Name Product Number
Alcalase 2.4 L Protease Bacillus licheniformis P4860
AMG 300 L Amyloglucosidase from Aspergillus niger A7095
BAN 240 L α-Amylase Bacillus amyloliquefaciens A7595
Carezyme 1000 L Cellulase from Aspergillus sp. C2605
Celluclast 1.5 L Cellulase from Trichoderma reesei ATCC 26921 C2730
Dextranase Plus L Dextranase Chaetomium erraticum D0443
Duramyl 300 α-Amylase Bacillus sp. A7720
Esperase 8.0 L Protease Bacillus sp. P5860
Everlase 16.0 L Protease Bacillus sp. P5985
Flavourzyme 500 L Protease from Aspergillus oryzae P6110
Fructozyme L Inulinase Aspergillus niger I2017
Fungamyl 800 L α-Amylase from Aspergillus oryzae A8220
Glucanex Lysing Enzymes from Trichoderma harzianum L1412
Lactozyme 3000 L β-Galactosidase Kluyveromyces lactis G3665
Lecitase 10 L Phospholipase A2 porcine pancreas P0861
Lecitase Ultra Phospholipase A1 from Thermomyces lanuginosus expressed in Aspergillus L3295
Lipolase 100 L Lipase from Thermomyces lanuginosus L0777
Neutrase 0.8 L Protease from Bacillus amyloliquefaciens P1236
Novozyme 188 Cellobiase from Aspergillus niger C6105
Novozyme 435 Lipase acrylic resin from Candida antarctica L4777
Novozymes CALA L Lipase from Candida expressed in Aspergillus L3420
Novozymes CALB L Lipase from Candida expressed in Aspergillus L3170
Palatase 20,000 L Lipase from Rhizomucor miehei L4277
Pectinex 3X L Pectinase from Aspergillus niger P2736
Pectinex Ultra SPL Pectinase from Aspergillus aculeatus P2611
Pentopan Mono BG Xylanase from Thermomyces lanuginosus X2753
Promozyme 400 L Pullulanase from Bacillus acidopullulyticus P2986
Protamex Protease from Bacillus P0029
Savinase 16.0 L Protease Bacillus sp. P3111
Subtilisin A Proteinase Bacillus licheniformis P5380
Sweetzyme T Glucose Isomerase Streptomyces murinus G4166
Termamyl 120 α-Amylase Bacillus licheniformis A3403
Viscozyme L Viscozyme L V2010
Bulk inquiries should be made directly to Novozymes Corporation at 1-800-879-6686 or online at novozymes.com
All products listed are registered trademarks of Novozymes™