Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Metabolomics > Enzyme Explorer > Application Index > Immuno Labelling
Enzyme Explorer
Immuno Labelling
  Alkaline Phosphatase
Peroxidase
Product Name Product # 
Alkaline Phosphatase  
Phosphatase, Alkaline bovine, minimum 4000 units/mg protein, recombinant, expressed in Pichia pastoris P8361
Phosphatase, Alkaline, biotinamidocaproyl labeled from calf intestine, Lyophilized powder, minimum 3,000 DEA units/mg protein P1318
Phosphatase, Alkaline from human placenta, minimum 1 unit/mg solid (in glycine buffer) P1391
Phosphatase, Alkaline-maleimide activated from calf intestine, Lyophilized powder, minimum 2,000 DEA units/mg protein P1959
Phosphatase, Alkaline from rabbit intestine, 0.2-0.6 unit/mg solid (in glycine buffer) P2265
Phosphatase, Alkaline bovine intestinal mucosa, Buffered aqueous glycerol solution, 4,000-6,000 DEA units/mg protein P3681
Phosphatase, Alkaline, polymerized and maleimide activated from calf intestine, Lyophilized powder, 1,200-2,400 DEA units/mg protein P3833
Phosphatase, Alkaline from human placenta, approx. 15 units/mg solid (in glycine buffer) P3895
Phosphatase, Alkaline from porcine intestinal mucosa, approx. 1 unit/mg solid (in glycine buffer) P4002
Phosphatase, Alkaline Escherichia coli, Buffered aqueous glycerol solution P4069
Phosphatase, Alkaline Escherichia coli, Suspension P4252
Phosphatase, Alkaline Escherichia coli, Ammonium sulfate suspension, approx. 10 units/mg protein (modified Warburg-Christian) P4377
Phosphatase, Alkaline from porcine kidney, Lyophilized powder, 100-300 DEA units/mg protein P4439
Phosphatase, Alkaline bovine kidney, Lyophilized powder P4653
Phosphatase, Alkaline from bovine milk, minimum 1 unit/mg solid (in glycine buffer) P5395
Phosphatase, Alkaline bovine intestinal mucosa, Type VII-S, Ammonium sulfate suspension, 2,000-4,000 DEA units/mg protein P5521
Phosphatase, Alkaline Escherichia coli, Lyophilized powder, 30-60 units/mg protein (in glycine buffer) P5931
Phosphatase, Alkaline bovine intestinal mucosa, Lyophilized powder, 2,000-4,000 DEA units/mg protein P6772
Phosphatase, Alkaline bovine intestinal mucosa, Buffered aqueous solution, 2,000-4,000 DEA units/mg protein P6774
Phosphatase, Alkaline from bovine liver, Lyophilized powder, approx. 10 units/mg protein (in glycine buffer) P7034
Phosphatase, Alkaline bovine intestinal mucosa, 10-30 DEA units/mg solid P7640
Phosphatase, Alkaline calf intestine, Buffered aqueous glycerol solution, minimum 4,000 DEA units/mg protein P7923
Phosphatase, Alkaline from canine intestine, 1-3 units/mg solid (in glycine buffer) P8639
Phosphatase, Alkaline-maleimide activated from calf intestine, Lyophilized powder, minimum 2,000 DEA units/mg protein P1959
Phosphatase, Alkaline, polymerized and maleimide activated from calf intestine, Lyophilized powder, 1,200-2,400 DEA units/mg protein P3833
Phosphatase alkaline bovine intestinal mucosa, BioChemika, solution (clear), >10000 U/ml 79385
Phosphatase alkaline calf intestinal mucosa, BioChemika, solution in 50% glycerol with 5 mM MgCl2 and 0.1 mM ZnCl2, pH ~7.0, ~4200 U/mg protein (10-20 mg/ml) 79396
Phosphatase alkaline calf intestinal mucosa, BioChemika, for enzyme immunoassay, solution (clear, colorless), ~2500 U/mg protein (~10 mg/ml) 79390
Phosphatase alkaline calf intestinal mucosa, BioChemika, suspension (aqueous), white, ~2000 U/mg protein (~10 mg/ml) 79393
Phosphatase alkaline calf intestinal mucosa, BioChemika, lyophilized, powder, 0.5-2.5 U/mg 79392
Phosphatase alkaline from E. coli, BioChemika, suspension, white, ~50 U/mg protein (~5-~10 mg/ml) 79377
Peroxidase back to top
Peroxidase horseradish, Essentially salt-free, lyophilized powder, 70-100 units/mg solid P0889
Peroxidase horseradish, Essentially salt-free, lyophilized powder, minimum 70 units/mg solid P1014
Peroxidase from Glycine max (soybean), Essentially salt-free, lyophilized powder, 50-150 units/mg solid P1432
Peroxidase, Maleimide Activated from horseradish, Lyophilized powder, 200-320 units/mg protein P1709
Peroxidase Polymerized and Maleimide Activated from horseradish, Lyophilized powder, 125-250 units/mg protein P1834
Peroxidase Highly stabilized from horseradish, Essentially salt-free, lyophilized powder, 200-300 units/mg solid P2088
Peroxidase from Arthromyces ramosus, Lyophilized powder, 50-200 units/mg solid P4794
Peroxidase from Arthromyces ramosus, BioChemika, powder, light brown, >400 U/mg 41366
Peroxidase Acidic Isoenzyme from horseradish, Type X, Ammonium sulfate suspension, approx. 250 units/mg protein (Biuret) P6140
Peroxidase horseradish, Essentially salt-free, lyophilized powder, 250-330 units/mg solid (using pyrogallol), Type VI-A P6782
Peroxidase horseradish, Type I, Essentially salt-free, lyophilized powder, 50-150 units/mg solid P8125
Peroxidase horseradish, Type II, Essentially salt-free, lyophilized powder, 150-250 units/mg solid P8250
Peroxidase horseradish, Type VI, Essentially salt-free, lyophilized powder, 250-330 units/mg solid P8375
Peroxidase horseradish, Type VI, Essentially salt-free, lyophilized powder, 250-330 units/mg solid P8375
Peroxidase horseradish, Type XII, Essentially salt-free, lyophilized powder, 250-330 units/mg solid P8415
Peroxidase from horse radish, BioChemika, lyophilized, powder, red-brown, ~850 U/mg 77334
Peroxidase from horse radish, BioChemika, lyophilized, powder, red-brown, ~700 U/mg 77333
Peroxidase from horse radish, BioChemika, powder, red-brown, ~550 U/mg 77335
Peroxidase from horse radish, BioChemika, lyophilized, powder, beige, ~150 U/mg 77332
Peroxidase from horse radish, BioChemika, essentially salt free, lyophilized, powder, light brown, ~100 U/mg 77330
  back to top