Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Enzyme Explorer
Cox Enzymes
Cyclooxygenase (COX) produces the key cyclic endoperoxide intermediate in the arachidonic acid cascade. The two COX isoforms are the constitutive form (COX1) and the inducible form (COX2). Many of the COX inhibitors are classic analgesics and non-steroidal anti-inflammatory drugs (NSAIDs). COX inhibitors have recently been implicated in cancer prevention and the slowing of atherosclerosis.
Product Product No.
Two New Cyclooxygenase Enzymes!
Cyclooxygenase 1, Sheep C 0733
Cyclooxygenase 2, Human Recombinant C 0858
Cyclooxygenase Inhibitors
N-(4-Acetamidophenyl)-indomethacin amide A 1224
Acetylsalicylic acid A 5376
5-Aminosalicylic acid A 3537
N-t-Butyl-alpha-phenylnitrone B 7263
DuP-697 D 4938
Diclofenac sodium D 6899
5,8,11,14-Eicosatetraynoic acid E 1768
Flurbiprofen F 8514
S(+)-Ibuprofen I-106
Ibuprofen I 4883
Indomethacin heptyl ester solution I 7904
Indomethacin I 7378
Ketorolac Tris salt K 1136
Ketoprofen K 1751
S(+)-Ketoprofen K 2135
(S)-6-Methoxy-alpha-methyl-2-naphthaleneacetic acid Na M 1275
(S)-6-Methoxy-alpha-methyl-2-naphthaleneacetic acid M 4015
Meloxicam M 3935
Niflumic acid N 0630
Nimesulide N 1016
NS-398 N-194
Nordihydroguaiaretic acid N 5023
N-(3-Pyridyl)indomethacinamide P 0865
Piroxicam P 5654
N-(2-Phenylethyl)indomethacinamide P 0740
Phenylbutazone P 8386
Resveratrol R 5010
Sulindac S 8139