Sigma-Aldrich
Life Science > Metabolomics > Enzyme Explorer > Cell Signaling Enzymes > Elafin
Enzyme Explorer
Elafin
Elafin
Human, Recombinant
Expressed in Saccharomyces cerevisiae
Product Code E7280
Synonyms: Elastase-specific inhibitor; Skin-Derived Antileukoprotease
Calculated Molecular Masses
Average Molecular Mass: 5999.20 Da
Monoisotopic Molecular Mass: 5994.8158 Da
Isoelectric Point (pI): 9.7
Elafin-fixed
Elafin Peptide Sequence1
Elafin is a peptide found in the human epithelia. It is a reversible, tightly binding inhibitor of human leukocyte elastase and proteinase 3 found in human epithelia.
Ki = 1.7 x 10-10 M for human leukocyte elastase2
Ki = 4.2 x 10-10 M for human proteinase 33
Elafin is produced in human skin, lungs, and mammary glands. It may affect the inflammation process and protect cells against destruction by the immune system. Elafin has shown potential applications for the treatment of cardiac infarction and organ rejection, as well as for treatment of seriously injured tissues and inflammatory lung diseases.4
Preparation Instructions
Stock solutions of Elafin may be prepared in water. Subsequent dilutions should be made into aqueous buffers, pH 4.0-8.0.
Storage/Stability
This product ships at ambient temperature and storage at 0 to -20°C is recommended. Stock solutions should be stored at -20°C. Avoid repeated freezing and thawing.
Related Products
Product Name Product # 
Elastase from human leukocytes E8140
Proteinase 3 from human leukocytes P0615
References
  1. Wiedow, O., et. al., J. Biol. Chem., 266, 3356 (1991)
  2. Ying, Q.L., et. al., Biochemistry, 32, 1866 (1991)
  3. Ying, Q.L., et. al., J. Respir. Cell. Mol. Biol., 24, 83 (2001)
  4. Sallenave, J.M., Respir. Res., 1, 87-92 (2000)
Browse Related Categories:
Elafin
Antibiotics
Enzyme Inhibitors
back to top
back to Enzyme Explorer