Enzyme Explorer
Class Index
|
|
|
| |
Sigma offers over 1,600 enzymes as well as associated inhibitors and substrates.
TO SEARCH FOR AN ENZYME BY EC # OR REACTION CATALYZED... 1. Click on a category in the menu below 2. Scroll down the list to the enzyme of interest 3. Click on the Product # to access product listing and related information including: inhibitors, substrates, assay procedures, Product Information Sheets, C of A and MSDS |
| |
1.x.x.x Oxidoreductases 2.x.x.x Transferases 3.x.x.x Hydrolases (Acting on esters) 3.x.x.x Hydrolases (Glycosidases) 3.x.x.x Hydrolases (Other) 4.x.x.x Lyases 5.x.x.x Isomerases 6.x.x.x Ligases
|
| |
Oxidoreductases |
|
| E.C. # |
Product Name |
Product # |
|
Acting on hydroxyl groups |
|
| 1.1.1.1 |
Alcohol Dehydrogenase from Saccharomyces cerevisiae, suitable for conventional determination of b-NAD, b-NADH, ethanol and acetaldehyde, Lyophilized powder, Solid with buffer salts, 300-500 units/mg protein |
A7011 |
| 1.1.1.1 |
Alcohol Dehydrogenase from Saccharomyces cerevisiae, suitable for recycling micro-assay of b-NAD and b-NADH, Lyophilized powder, Solid with buffer salts, minimum 300 units/mg protein |
A3263 |
| 1.1.1.1 |
Alcohol dehydrogenase–Agarose from Baker's yeast (S. cerevisiae), Lyophilized powder, 900-1,200 units/g agarose, pH 8.8, 30°C |
A2529 |
| 1.1.1.1 |
Alcohol Dehydrogenase equine liver, Lyophilized powder, ³0.5 units/mg protein (TCA/Biuret) |
A9589 |
| 1.1.1.1 |
Alcohol Dehydrogenase from Saccharomyces cerevisiae, BioChemika, powder, off-white, ³250 U/mg |
05635 |
| 1.1.1.1 |
Alcohol Dehydrogenase from Saccharomyces cerevisiae, BioChemika, lyophilized, powder, ~300 U/mg |
05640 |
| 1.1.1.1 |
Alcohol Dehydrogenase equine liver, BioChemika, suspension in 20 mM potassium phosphate buffer, pH 7; ethanol 10%, ampoule, 10% as ethanol, ~30 U/ml |
05645 |
| 1.1.1.1 |
Alcohol Dehydrogenase equine liver, BioChemika, crystalline suspension in 20 mM potassium phosphate buffer, ~3 U/mg protein (~10 mg/ml) |
05646 |
| 1.1.1.1 |
Alcohol Dehydrogenase equine liver, BioChemika, lyophilisate, slightly beige, ³0.25 U/mg |
17233 |
| 1.1.1.1 |
Alcohol Dehydrogenase from Candida boidinii, BioChemika, powder, light brown, ³0.4 U/mg |
91031 |
| 1.1.1.2 |
Alcohol dehydrogenase, NADP+ dependent Thermoanaerobium brockii, Lyophilized powder, 30-90 units/mg protein |
A9287 |
| 1.1.1.2 |
Alcohol Dehydrogenase Thermoanaerobium brockii, Lyophilized powder, 5-15 units/mg protein |
A8435 |
| 1.1.1.2 |
(S)-Aromatic Alcohol Dehydrogenase, NADP+ dependent from Thermoanaerobium sp., Lyophilized powder, 1-5 units/mg solid |
A9685 |
| 1.1.1.2 |
Alcohol dehydrogenase, NADP+ dependent Thermoanaerobium brockii, Lyophilized powder, 30-90 units/mg protein |
A9287 |
| 1.1.1.2 |
Alcohol Dehydrogenase Thermoanaerobium brockii, Lyophilized powder, 5-15 units/mg protein |
A8435 |
| 1.1.1.2 |
(S)-Aromatic Alcohol Dehydrogenase, NADP+ dependent from Thermoanaerobium sp., Lyophilized powder, 1-5 units/mg solid |
A9685 |
| 1.1.1.2 |
Alcohol Dehydrogenase from Lactobacillus kefir, BioChemika, powder, beige, ~0.4 U/mg |
05643 |
| 1.1.1.2 |
Alcohol Dehydrogenase Thermoanaerobium brockii, BioChemika, powder, white, ~35 U/mg |
05655 |
| 1.1.1.6 |
Glycerol Dehydrogenase from Bacillus megaterium, Lyophilized powder, ³15 units/mg solid, pH 8.8 |
G6267 |
| 1.1.1.6 |
Glycerol Dehydrogenase from Cellulomonas sp., Lyophilized powder, 50-125 units/mg protein, pH 10.0 |
G3512 |
| 1.1.1.6 |
Glycerol Dehydrogenase Enterobacter aerogenes, Lyophilized powder, 20-80 units/mg protein, pH 10.0 |
G4783 |
| 1.1.1.6 |
Glycerol Dehydrogenase Geotrichum candidum, BioChemika, ~30 U/g |
49860 |
| 1.1.1.8 |
a-Glycerophosphate Dehydrogenase from rabbit muscle, Type I, Ammonium sulfate suspension, 100-300 units/mg protein |
G6751 |
| 1.1.1.8 |
a-Glycerophosphate Dehydrogenase from rabbit muscle, Type X, Lyophilized powder, 100-300 units/mg protein |
G6880 |
| 1.1.1.8 |
sn-Glycero-3-phosphate Dehydrogenase rabbit muscle, BioChemika, suspension, white, ~270 U/mg protein (~10 mg/ml) |
50013 |
| 1.1.1.8 |
sn-Glycero-3-phosphate Dehydrogenase rabbit muscle, BioChemika, lyophilized, powder, white, 100-250 U/mg |
50015 |
| 1.1.1.10 |
L-Xylulose Reductase from pigeon liver, Ammonium sulfate suspension, 5-15 units/mg protein (BCA) |
X5250 |
| 1.1.1.14 |
Sorbitol Dehydrogenase from sheep liver, Lyophilized powder, minimum 20 units/mg protein |
S3764 |
| 1.1.1.14 |
Sorbitol Dehydrogenase from sheep liver, 20-40 units/mg protein |
S1128 |
| 1.1.1.14 |
SDH Control (Sorbitol Dehydrogenase) |
S8135 |
| 1.1.1.14 |
Sorbitol Dehydrogenase from sheep liver, BioChemika, lyophilized, powder, white, ~6 U/mg |
85535 |
| 1.1.1.18 |
myo-Inositol Dehydrogenase from Enterobacter aerogenes, Lyophilized powder, 25-50 units/mg protein |
I5010 |
| 1.1.1.18 |
myo-Inositol Dehydrogenase from Enterobacter aerogenes, Lyophilized powder, 10-20 units/mg protein |
I0255 |
| 1.1.1.18 |
myo-Inositol Dehydrogenase from Enterobacter aerogenes, 10-20 units/mL, Ammonium sulfate suspension |
I3517 |
| 1.1.1.22 |
Uridine-5¢-diphosphoglucose Dehydrogenase from bovine liver, Type III, Lyophilized powder, 0.02-0.08 units/mg protein |
U5500 |
| 1.1.1.22 |
Uridine-5¢-diphosphoglucose Dehydrogenase from bovine liver, Type VI, Lyophilized powder, 0.3-0.6 units/mg protein |
U7251 |
| 1.1.1.26 |
Glyoxylate reductase from Spinacia oleracea (spinach), Ammonium sulfate suspension, approx. 50 units/mg protein (Biuret) |
G5259 |
| 1.1.1.27 |
L-Lactic Dehydrogenase LDH-5 (M4) Isoenzyme solution from bovine muscle, Type XXVI, Ammonium sulfate suspension, minimum 600 units/mg protein |
L0508 |
| 1.1.1.27 |
L-Lactic Dehydrogenase LDH-5 (M4) Isoenzyme solution from porcine muscle, Type XXXII-S, Ammonium sulfate suspension, approx. 1,000 units/mg protein |
L5762 |
| 1.1.1.27 |
L-Lactic Dehydrogenase LDH-2 (H3M) Isoenzyme from human erythrocytes, Type XX, 98% (homogeneous), Ammonium sulfate suspension, approx. 150 units/mg protein |
L3757 |
| 1.1.1.27 |
L-Lactic Dehydrogenase solution porcine heart, Ammonium sulfate suspension, minimum 400 units/mg protein (Biuret) |
L7525 |
| 1.1.1.27 |
L-Lactic Dehydrogenase Bacillus stearothermophilus, recombinant, expressed in Escherichia coli, Lyophilized powder, minimum 200 units/mg protein, pH 6.0, 30°C (Lowry) |
L3151 |
| 1.1.1.27 |
L-Lactic Dehydrogenase from bovine muscle, Type X, Ammonium sulfate suspension, minimum 600 units/mg protein |
L1378 |
| 1.1.1.27 |
L-Lactic Dehydrogenase isoenzymes LDH-1 (H4) and LDH-2 (H3M) from human erythrocytes, Type XII, Ammonium sulfate suspension, 300-500 units/mg protein |
L3379 |
| 1.1.1.27 |
L-Lactic Dehydrogenase from rabbit muscle, Type XI, Salt-free, lyophilized powder, 700-1,200 units/mg protein |
L1254 |
| 1.1.1.27 |
L-Lactic Dehydrogenase solution from rabbit muscle, Type I, Ammonium sulfate suspension, 40-100 units/mg protein |
L2375 |
| 1.1.1.27 |
L-Lactic Dehydrogenase solution from rabbit muscle, Type II, Ammonium sulfate suspension, 800-1,200 units/mg protein |
L2500 |
| 1.1.1.27 |
L-Lactic Dehydrogenase solution from rabbit muscle, Type V-S, >95%, Ammonium sulfate suspension, minimum 700 units/mg protein |
L5132 |
| 1.1.1.27 |
L-Lactic dehydrogenase Insoluble enzyme attached to beaded agarose from rabbit muscle, Ammonium sulfate suspension, 300-500 units/g agarose |
L8011 |
| 1.1.1.27 |
L-Lactic dehydrogenase Insoluble enzyme attached to polyacrylamide from rabbit muscle, 25-40 units/g solid |
L8253 |
| 1.1.1.27 |
L-Lactic Dehydrogenase LDH-1 (H4) Isoenzyme solution from rabbit heart, Type XXXVI, Ammonium sulfate suspension, 400-600 units/mg protein |
L2889 |
| 1.1.1.27 |
L-Lactic Dehydrogenase from chicken heart, Type VIII, Ammonium sulfate suspension, 500-800 units/mg protein |
L9126 |
| 1.1.1.27 |
L-Lactic Dehydrogenase solution bovine heart, Type III, Ammonium sulfate suspension, 500-700 units/mg protein |
L2625 |
| 1.1.1.27 |
L-Lactic Dehydrogenase solution bovine heart, Type IX, Ammonium sulfate suspension, minimum 400 units/mg protein |
L0377 |
| 1.1.1.27 |
L-Lactic Dehydrogenase solution bovine heart, Type XVII, Buffered aqueous glycerol solution, 400-600 units/mg protein |
L1006 |
| 1.1.1.27 |
L-Lactic Dehydrogenase solution from rabbit muscle, Type XXXIX, Buffered aqueous glycerol solution, 700-1,000 units/mg protein |
L2518 |
| 1.1.1.27 |
L-Lactate Dehydrogenase rabbit muscle, BioChemika, in 2.1 M ammonium sulfate solution, pH ~6.0, suspension, white, ~500 U/mg protein (~10 mg/ml) |
61311 |
| 1.1.1.27 |
L-Lactate Dehydrogenase rabbit muscle, BioChemika, lyophilized, powder, off-white, ~140 U/mg |
61309 |
| 1.1.1.27 |
Lactic Dehydrogenase preparation from bovine heart, BioChemika, off-white, ~250 U/mg protein (~10 mg/ml) |
61310 |
| 1.1.1.28 |
D-Lactic Dehydrogenase Lactobacillus leichmanii, Lyophilized powder, 150-300 units/mg protein |
L3888 |
| 1.1.1.28 |
D-Lactic Dehydrogenase solution from Lactobacillus leichmanii, Ammonium sulfate suspension, 250-500 units/mg protein (Biuret) |
L2011 |
| 1.1.1.28 |
D-Lactic Dehydrogenase Leuconostoc mesenteroides, Ammonium sulfate suspension, 1,000-3,000 units/mg protein (Biuret) |
L2395 |
| 1.1.1.28 |
D-Lactic Dehydrogenase from Staphylococcus epidermidis, Lyophilized powder, minimum 10 units/mg solid |
L9636 |
| 1.1.1.28 |
D-Lactate Dehydrogenase Staphylococci, BioChemika, lyophilisate, white, ~120 U/mg |
17847 |
| 1.1.1.28 |
D-Lactate Dehydrogenase Lactobacillus sp., BioChemika, powder, white, ³400 U/mg |
59023 |
| 1.1.1.28 |
D-Lactic Dehydrogenase Lactobacillus leichmanii, BioChemika, in 3.2 M ammonium sulfate solution, pH ~6.0, suspension, white, ~1000 U/ml |
61306 |
| 1.1.1.30 |
b-Hydroxybutyrate Dehydrogenase Rhodopseudomonas sphaeroides, Type V, Lyophilized powder, 250-750 units/mg protein |
H8509 |
| 1.1.1.30 |
b-Hydroxybutyrate Dehydrogenase from Rhodopseudomonas sphaeroides, Type II, Ammonium sulfate suspension, 5-10 units/mg protein (Biuret) |
H6126 |
| 1.1.1.30 |
b-Hydroxybutyrate Dehydrogenase from Pseudomonas lemoignei, Lyophilized powder, 200-750 units/mg protein |
H9408 |
| 1.1.1.30 |
b-Hydroxybutyrate Dehydrogenase from Rhodopseudomonas sphaeroides, Type III, Ammonium sulfate suspension, minimum 20 units/mg protein (Biuret) |
H4005 |
| 1.1.1.30 |
b-Hydroxybutyrate Dehydrogenase from Pseudomonas lemoignei, 50-100 units/mL, Ammonium sulfate suspension |
H6523 |
| 1.1.1.30 |
b-Hydroxybutyrate Dehydrogenase Rhodopseudomonas sphaeroides, BioChemika, in 3.2 M ammonium sulfate solution, pH ~6.0, suspension, white, 4-10 U/mg protein (~5 mg/ml) |
54975 |
| 1.1.1.37 |
Malic Dehydrogenase from Thermus flavus, Lyophilized powder, minimum 50 units/mg solid |
M7032 |
| 1.1.1.37 |
Malic Dehydrogenase porcine heart, Buffered aqueous glycerol solution, approx. 1,000 units/mg protein (Biuret) |
M2634 |
| 1.1.1.37 |
Malic Dehydrogenase porcine heart, Ammonium sulfate suspension, approx. 400 units/mg protein (Biuret) |
M7383 |
| 1.1.1.37 |
Malic Dehydrogenase from bovine heart, Ammonium sulfate suspension, 2000-4000 units/mg protein (modified Warburg-Christian) |
M9004 |
| 1.1.1.37 |
Malic Dehydrogenase Thermus sp., recombinant, expressed in Escherichia coli, Ammonium sulfate suspension, 100-200 units/mg protein (Lowry) |
M1035 |
| 1.1.1.37 |
Malic Dehydrogenase porcine heart, BioChemika, suspension, slightly beige, ~1200 U/mg protein (~5 mg/ml) |
63170 |
| 1.1.1.42 |
Isocitrate Dehydrogenase preparation porcine heart, Type I |
I1877 |
| 1.1.1.42 |
Isocitric Dehydrogenase (NADP) solution porcine heart, Type IV, Buffered aqueous glycerol solution, 3-20 units/mg protein (Biuretapprox. 0.1% of that activity is obtained with NAD as cofactor.) |
I2002 |
| 1.1.1.42 |
1,3-Indandione, 97% |
I-200-2 |
| 1.1.1.42 |
Isocitrate Dehydrogenase preparation porcine heart, Lyophilized powder, 20-60 units/mg protein (Biuret) |
I2516 |
| 1.1.1.42 |
Isocitrate Dehydrogenase preparation porcine heart, BioChemika, shipped in dry ice, powder, yellow-brown, 4-12 U/mg |
58774 |
| 1.1.1.42 |
Isocitrate Dehydrogenase preparation porcine heart, BioChemika, solution (clear), amber, 3-8 U/mg protein (~12 mg/ml) |
58775 |
| 1.1.1.44 |
6-Phosphogluconic Dehydrogenase from Lactobacillus sp., Lyophilized powder |
P0235 |
| 1.1.1.44 |
6-Phosphogluconic Dehydrogenase from Saccharomyces cerevisiae, Lyophilized powder, 3-6 units/mg solid |
P4553 |
| 1.1.1.44 |
6-Phosphogluconic Dehydrogenase from torula yeast, Type IV, Ammonium sulfate suspension, minimum 20 units/mg protein |
P0507 |
| 1.1.1.44 |
6-Phosphogluconic Dehydrogenase from sheep liver, Lyophilized powder, 2-10 units/mg protein |
P8406 |
| 1.1.1.44 |
6-Phosphogluconic Dehydrogenase from torula yeast, BioChemika, ³15 U/mg protein |
79461 |
| 1.1.1.47 |
Glucose Dehydrogenase Bacillus megaterium, Lyophilized powder, 50-150 units/mg solid |
G7653 |
| 1.1.1.47 |
Glucose Dehydrogenase from Thermoplasma acidophilum, recombinant, expressed in Escherichia coli, Ammonium sulfate suspension, minimum 100 units/mg protein (glucose substrate), minimum 50 units/mg protein (galactose substrate), Solution |
G5909 |
| 1.1.1.47 |
Glucose Dehydrogenase Bacillus megaterium, BioChemika, ~33 U/mg |
49165 |
| 1.1.1.48 |
b-Galactose Dehydrogenase solution from Pseudomonas fluorescens, recombinant, expressed in Escherichia coli, Ammonium sulfate suspension, minimum 50 units/mg protein (Biuret) |
G6637 |
| 1.1.1.49 |
Glucose-6-phosphate Dehydrogenase–Agarose from Baker's yeast (S. cerevisiae), Ammonium sulfate suspension, 1,000-1,750 units/g agarose, 30°C |
G5506 |
| 1.1.1.49 |
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, recombinant, expressed in Escherichia coli, Lyophilized powder, 550-1,100 units/mg protein (Biuret) |
G8529 |
| 1.1.1.49 |
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, recombinant, expressed in Escherichia coli, Ammonium sulfate suspension, 550-1,100 units/mg protein (Biuret) |
G8404 |
| 1.1.1.49 |
Glucose-6-phosphate Dehydrogenase from torula yeast, Ammonium sulfate suspension, 300-600 units/mg protein (Biuret) |
G8289 |
| 1.1.1.49 |
Glucose-6-phosphate Dehydrogenase Biotin Labeled from Baker's yeast, Type XV-B, Lyophilized powder, 80-160 units/mg protein |
G3386 |
| 1.1.1.49 |
Glucose-6-phosphate Dehydrogenase from baker's yeast (S. cerevisiae), Type XV, Lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian) |
G6378 |
| 1.1.1.49 |
Glucose-6-phosphate Dehydrogenase from baker's yeast (S. cerevisiae), Type IX, Lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian) |
G4134 |
| 1.1.1.49 |
Glucose-6-phosphate Dehydrogenase from baker's yeast (S. cerevisiae), Type VII, Ammonium sulfate suspension, 300-400 units/mg protein (modified Warburg-Christian) |
G7877 |
| 1.1.1.49 |
Glucose-6-phosphate Dehydrogenase from baker's yeast (S. cerevisiae), Type V, Dry ammonium sulfate cake, ³150 units/mg protein (modified Warburg-Christian) |
G7750 |
| 1.1.1.49 |
Glucose-6-phosphate Dehydrogenase from torula yeast, Lyophilized powder, 200-600 units/mg protein (BiuretContains approx. 20% sodium citrate based on dry weight.) |
G8164 |
| 1.1.1.49 |
Glucose-6-phosphate Dehydrogenase from yeast, BioChemika, lyophilized, powder, off-white, ~160 U/mg |
49273 |
| 1.1.1.49 |
Glucose-6-phosphate Dehydrogenase from yeast, BioChemika, standardized with bovine serum albumin, crystalline suspension in 3.2 M ammonium sulfate solution, pH ~6, ~120 U/mg protein (~5 mg/ml) |
49270 |
| 1.1.1.49 |
Glucose-6-phosphate Dehydrogenase from baker's yeast (S. cerevisiae), BioChemika, crystalline suspension in 3.2 M ammonium sulfate solution, pH ~6, ~240 U/mg protein (~1 mg/ml) |
49271 |
| 1.1.1.49 |
Glucose-6-phosphate Dehydrogenase from baker's yeast (S. cerevisiae), BioChemika, lyophilized, powder, white, ~210 U/mg |
49272 |
| 1.1.1.49 |
Glucose-6-phosphate Dehydrogenase from torula yeast, BioChemika, suspension, white, ~180 U/mg protein (~1 mg/ml) |
49279 |
| 1.1.1.49 |
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, BioChemika, lyophilized, powder, slightly beige, ~170 U/mg |
49275 |
| 1.1.1.50 |
3a-Hydroxysteroid Dehydrogenase preparation from Pseudomonas testosteroni, Lyophilized powder, 20-50 units/mg protein |
H1506 |
| 1.1.1.50 |
3a-Hydroxysteroid Dehydrogenase preparation from Pseudomonas testosteroni, 50-100 units/mL, Ammonium sulfate suspension |
H6398 |
| 1.1.1.50 |
3a-Hydroxysteroid Dehydrogenase from Pseudomonas testosteroni, BioChemika, lyophilized, powder, white, ~9 U/mg |
56460 |
| 1.1.1.67 |
Mannitol Dehydrogenase from Leuconostoc mesenteroides |
M9532 |
| 1.1.1.95 |
3-Phosphoglycerate Dehydrogenase from chicken liver, Lyophilized powder, 1.5-7.0 units/mg protein (modified Warburg-Christian) |
P4579 |
| 1.1.1.119 |
Glucose Dehydrogenase, NADP dependent from Cryptococcus uniguttulatus, Lyophilized powder, minimum 10 units/mg solid |
G0649 |
| 1.1.1.176 |
12a-Hydroxysteroid Dehydrogenase from Bacillus sphaericus, Lyophilized powder, 150-350 units/mg protein (Lowry) |
H2265 |
| 1.1.1.176 |
12a-Hydroxysteroid Dehydrogenase Clostridium sp., BioChemika, solution, yellow, ³18 U/ml |
37762 |
| 1.1.1.205 |
IMPDH |
I1782 |
| 1.1.1.233 |
N-Acylmannosamine 1-Dehydrogenase from Pseudomonas sp., Lyophilized powder, minimum 45 units/mg protein |
A7561 |
| 1.1.2.3 |
L-Lactic Dehydrogenase (cytochrome) from baker's yeast (S. cerevisiae), Type IV-SS, Ammonium sulfate suspension, 0.1-0.6 units/mg protein (Biuret) |
L4506 |
| 1.1.3.4 |
Glucose Oxidase Aspergillus niger, Type V-S, approx. 1,000 units/mL (without added oxygen), Buffered aqueous solution |
G6891 |
| 1.1.3.4 |
Glucose Oxidase Aspergillus niger, 2,000-10,000 units/g solid (without added oxygen) |
G6766 |
| 1.1.3.4 |
Glucose Oxidase Aspergillus niger, Type VII, Lyophilized powder, 100,000-200,000 units/g solid (without added oxygen) |
G2133 |
| 1.1.3.4 |
Glucose Oxidase Aspergillus niger, Type II, 15,000-25,000 units/g solid (without added oxygen) |
G6125 |
| 1.1.3.4 |
Glucose Oxidase Aspergillus niger, Type X-S, Lyophilized powder, 100,000-250,000 units/g solid (without added oxygen) |
G7141 |
| 1.1.3.4 |
Glucose Oxidase Aspergillus niger, Buffered aqueous solution, minimum 200 units/mg protein (Biuret) |
G9010 |
| 1.1.3.4 |
Glucose Oxidase Aspergillus niger, Type II-S, 25,000-50,000 units/g solid (without added oxygen) |
G6641 |
| 1.1.3.4 |
Glucose Oxidase Aspergillus niger, Type VII-S, Lyophilized powder, 100,000-250,000 units/g solid (without added oxygen) |
G7016 |
| 1.1.3.4 |
Glucose Oxidase Aspergillus niger, BioChemika, solution (clear), yellow, ~1000 U/ml |
49177 |
| 1.1.3.4 |
Glucose Oxidase Aspergillus niger, BioChemika, lyophilized, powder, ~200 U/mg |
49180 |
| 1.1.3.4 |
Glucose Oxidase Aspergillus niger, BioChemika, powder, yellow, ~150 U/mg |
49182 |
| 1.1.3.4 |
Glucose Oxidase Aspergillus niger, BioChemika, powder, yellow, ~130 U/mg |
49181 |
| 1.1.3.4 |
Glucose Oxidase Aspergillus niger, BioChemika, lyophilized, powder, ~25 U/mg |
49178 |
| 1.1.3.6 |
Cholesterol Oxidase from Pseudomonas fluorescens, Lyophilized powder, 10-50 units/mg protein |
C7149 |
| 1.1.3.6 |
Cholesterol Oxidase microbial, recombinant, expressed in Escherichia coli, Lyophilized powder |
C1235 |
| 1.1.3.6 |
Cholesterol Oxidase from Streptomyces sp., Lyophilized powder, minimum 25 units/mg protein |
C8649 |
| 1.1.3.6 |
Cholesterol Oxidase from Cellulomonas sp., Buffered aqueous solution, 20-60 units/mg protein (Biuret) |
C5421 |
| 1.1.3.6 |
Cholesterol Oxidase Nocardia erythropolis, BioChemika, in 1 M ammonium sulfate solution, pH 6, solution (slightly hazy), pale yellow, ³15 U/ml |
26746 |
| 1.1.3.6 |
Cholesterol Oxidase from Pseudomonas sp., BioChemika, powder, light yellow, >1 U/mg |
26747 |
| 1.1.3.9 |
Galactose Oxidase from Dactylium dendroides, Lyophilized powder, 500-1,500 units/mg protein |
G7907 |
| 1.1.3.9 |
Galactose Oxidase from Dactylium dendroides, Lyophilized powder, minimum 3,000 units/g solid |
G7400 |
| 1.1.3.10 |
Pyranose Oxidase from Cariolus sp., recombinant, expressed in Escherichia coli, 2.7 units/mg protein |
P4234 |
| 1.1.3.13 |
Alcohol Oxidase Candida boidinii, Lyophilized powder, 5-15 units/mg protein |
A6941 |
| 1.1.3.13 |
Alcohol Oxidase from Hansenula sp., Vacuum-dried powder, 20-40 units/mg protein (Bradford) |
A0438 |
| 1.1.3.13 |
Alcohol Oxidase solution from Pichia pastoris, BioChemika, solution, red to deep red, ³400 U/ml |
92705 |
| 1.1.3.15 |
Glycolate Oxidase Spinacia oleracea (spinach), Ammonium sulfate suspension, 3-12 units/mg protein, pH 7.8 (Biuret) |
G8260 |
| 1.1.3.17 |
Choline Oxidase from Alcaligenes sp., Lyophilized powder, approx. 10 units/mg solid |
C5896 |
| 1.1.3.17 |
Choline Oxidase from Alcaligenes sp., BioChemika, powder, yellow, 9-18 U/mg |
26986 |
| 1.1.3.17 |
Choline Oxidase from Arthrobacter globiformis, Lyophilized powder, 8-20 units/mg solid |
C4405 |
| 1.1.3.21 |
Glycerol 3-phosphate Oxidase from Pediococcus sp., Lyophilized powder, 40-80 units/mg solid, pH 8.1 |
G9637 |
| 1.1.3.21 |
Glycerol 3-phosphate Oxidase from Streptococcus thermophilus, Lyophilized powder, minimum 10 units/mg solid, pH 7.0 |
G4388 |
| 1.1.3.21 |
Glycerol 3-phosphate Oxidase from Aerococcus viridans, Lyophilized powder, minimum 70 units/mg solid, pH 8.1 |
G9888 |
| 1.1.3.21 |
Glycerol 3-phosphate Oxidase from Aerococcus viridans, BioChemika, powder, yellow, ~40 U/mg |
50030 |
| 1.1.3.22 |
Xanthine Oxidase from buttermilk, Lyophilized powder, 0.4-1.0 units/mg protein |
X4376 |
| 1.1.3.22 |
Xanthine Oxidase from buttermilk, Grade I, Ammonium sulfate suspension, approx. 0.5 units/mg protein |
X1875 |
| 1.1.3.22 |
Xanthine Oxidase from buttermilk, Grade III, Ammonium sulfate suspension, 1-2 units/mg protein |
X4500 |
| 1.1.3.22 |
Xanthine Oxidase microbial, Lyophilized powder, minimum 7 units/mg solid |
X2252 |
| 1.1.3.22 |
Xanthine Oxidase from buttermilk, Grade IV, Ammonium sulfate suspension, 0.1-0.4 units/mg protein |
X4875 |
| 1.1.3.22 |
Xanthine Oxidase from buttermilk, BioChemika, lyophilized, powder, brown, ~0.4 U/mg |
95493 |
| 1.1.3.29 |
N-Acylhexosamine Oxidase from Pseudomonas sp., Solution, minimum 20 units/mg protein |
A0186 |
| 1.1.99.11 |
D-Fructose Dehydrogenase from Gluconobacter industrius, Lyophilized powder, 400-1,200 units/mg protein |
F5152 |
| 1.1.99.11 |
D-Fructose Dehydrogenase from Gluconobacter industrius, Lyophilized powder, minimum 20 units/mg solid |
F4892 |
| 1.1.99.11 |
D-Fructose Dehydrogenase from Gluconobacter industrius, BioChemika, powder, light red, ³20 U/mg |
70052 |
|
Acting on aldehydes or oxo groups |
|
| 1.2.1.1 |
Formaldehyde Dehydrogenase (glutathione-dependent) from Candida boidinii, Lyophilized powder, 7-20 units/mg protein |
F4136 |
| 1.2.1.1 |
Formaldehyde Dehydrogenase (glutathione-dependent) from Candida boidinii, BioChemika, ~2.5 U/mg |
47584 |
| 1.2.1.1 |
Formate Dehydrogenase Pseudomonas spec., recombinant from E. coli, BioChemika, solution, light yellow, ~100 U/ml |
79900 |
| 1.2.1.2 |
Formate Dehydrogenase Candida boidinii, Lyophilized powder, 1.5-4.5 units/mg protein |
F5632 |
| 1.2.1.2 |
Formate Dehydrogenase Candida boidinii, Lyophilized powder, 5-15 units/mg protein |
F8649 |
| 1.2.1.2 |
Formate Dehydrogenase Pseudomonas spec., recombinant mutante to 79900 from E. coli, BioChemika, solution, light yellow, >150 U/ml |
75274 |
| 1.2.1.2 |
Formate Dehydrogenase Pseudomonas spec., recombinant mutante to 79900 NADP-dependant from E. coli from E. coli, BioChemika, solution, light yellow, ~50 U/ml |
79331 |
| 1.2.1.2 |
Formate Dehydrogenase Candida boidinii, BioChemika, powder, 0.3-0.6 U/mg |
47709 |
| 1.2.1.2 |
Formate Dehydrogenase Candida boidinii, BioChemika, liquid (clear), brown, ~50 U/ml |
47753 |
| 1.2.1.5 |
Aldehyde Dehydrogenase, potassium-activated baker's yeast (S. cerevisiae), Lyophilized powder, 2-10 units/mg protein |
A6338 |
| 1.2.1.12 |
Glyceraldehyde-3-phosphate Dehydrogenase human erythrocytes, Lyophilized powder, 50-150 units/mg protein |
G6019 |
| 1.2.1.12 |
Glyceraldehyde-3-phosphate Dehydrogenase from chicken muscle, Lyophilized powder, 40-100 units/mg protein |
G9263 |
| 1.2.1.12 |
Glyceraldehyde-3-phosphate Dehydrogenase from rabbit muscle, Ammonium sulfate suspension, 80-120 units/mg protein (Biuret) |
G0763 |
| 1.2.1.12 |
Glyceraldehyde-3-phosphate Dehydrogenase from rabbit muscle, Lyophilized powder, approx. 80 units/mg protein |
G2267 |
| 1.2.1.12 |
Glyceraldehyde-3-phosphate Dehydrogenase from Baker's yeast (S. cerevisiae), Lyophilized powder, 70-140 units/mg protein |
G5537 |
| 1.2.1.12 |
Glyceraldehyde-3-phosphate Dehydrogenase Agarose from Baker's yeast (S. cerevisiae), Lyophilized powder, 50-150 units/g dry agarose, pH 7.6, 30°C |
G0389 |
| 1.2.1.12 |
Glyceraldehyde-3-phosphate Dehydrogenase from rabbit muscle, BioChemika, suspension, white, ³50 U/mg protein (~10 mg/ml) |
49830 |
| 1.2.1.16 |
Succinic-Semialdehyde Dehydrogenase from Pseudomonas fluorescens, Lyophilized powder, 10-30 units/mg protein |
S3907 |
| 1.2.1.46 |
Formaldehyde Dehydrogenase from Pseudomonas putida, Lyophilized powder, 1-6 units/mg solid |
F1879 |
| 1.2.3.3 |
Pyruvate Oxidase bacterial, Lyophilized powder, minimum 1.5 units/mg |
P4591 |
| 1.2.3.3 |
Pyruvate Oxidase from Aerococcus sp., Lyophilized powder, minimum 35 units/mg protein (Biuret) |
P4105 |
| 1.2.3.4 |
Oxalate Oxidase from barley seedlings, Lyophilized powder, 0.5-1.5 units/mg solid |
O4127 |
| 1.2.4.2 |
a-Ketoglutarate Dehydrogenase from porcine heart, Buffered aqueous glycerol solution, 0.1-1 units/mg protein (Lowry) |
K1502 |
|
Acting on CH-CH |
|
| 1.3.3.5 |
Bilirubin Oxidase from Myrothecium verrucaria, Lyophilized powder, 15-65 units/mg protein |
B0390 |
| 1.3.3.6 |
Acyl coenzyme A oxidase from Arthrobacter sp., Lyophilized powder, 10-40 units/mg protein |
A2167 |
| 1.3.3.6 |
Acyl coenzyme A oxidase from Candida sp., Lyophilized powder, 10-30 units/mg protein |
A2903 |
|
Acting on CH-NH2 |
|
| 1.4.1.1 |
L-Alanine Dehydrogenase Bacillus subtilis, Buffered aqueous glycerol solution, approx. 30 units/mg protein (Lowry) |
A7653 |
| 1.4.1.1 |
L-Alanine Dehydrogenase Bacillus subtilis, Ammonium sulfate suspension, approx. 40 units/mg protein (Lowry) |
A7189 |
| 1.4.1.1 |
L-Alanine Dehydrogenase Bacillus subtilis, BioChemika, in 2.4 M ammonium sulfate solution, pH 7.0, suspension, white, ³30 U/mg protein (~10 mg/ml) |
05192 |
| 1.4.1.3 |
L-Glutamic Dehydrogenase solution from bovine liver, Type I, Ammonium sulfate suspension, approx. 40 units/mg protein |
G2501 |
| 1.4.1.3 |
L-Glutamic Dehydrogenase solution from bovine liver, Type II, Buffered aqueous glycerol solution, approx. 40 units/mg protein |
G2626 |
| 1.4.1.3 |
Glutamate Dehydrogenase bovine liver, Type III, Lyophilized powder, approx. 25 units/mg protein |
G7882 |
| 1.4.1.3 |
Glutamate Dehydrogenase bovine liver, BioChemika, solution (viscous), amber, ~1600 U/ml |
49390 |
| 1.4.1.3 |
Glutamate Dehydrogenase bovine liver, BioChemika, lyophilized, powder, >30 U/mg |
49392 |
| 1.4.1.4 |
L-Glutamic Dehydrogenase (NADP) from Proteus sp., Buffered aqueous solution, minimum 400 units/mg protein (Biuret) |
G4387 |
| 1.4.1.4 |
L-Glutamic Dehydrogenase (NADP) from Candida utilis, Lyophilized powder, 50-200 units/mg protein |
G5027 |
| 1.4.1.4 |
L-Leucine Dehydrogenase from Bacillus cereus, BioChemika, solution, slightly brown, ~50 U/ml |
40453 |
| 1.4.1.20 |
L-Phenylalanine Dehydrogenase from Sporosarcina sp., Lyophilized powder, minimum 6 units/mg solid |
P4798 |
| 1.4.1.20 |
L-Phenylalanine Dehydrogenase Rhodococcus sp. M4, BioChemika, solution (clear), light yellow, ~150 U/ml |
55001 |
| 1.4.3.2 |
L-Amino Acid Oxidase from Crotalus durissus venom, Ammonium sulfate suspension, 3-8 units/mg protein |
A2805 |
| 1.4.3.2 |
L-Amino Acid Oxidase from Crotalus adamanteus, Type I, Dried venom, ³0.3 unit/mg solid |
A9253 |
| 1.4.3.2 |
L-Amino Acid Oxidase Bothrops atrox venom, Type II, Dried venom, minimum 0.2 unit/mg solid |
A4257 |
| 1.4.3.2 |
L-Amino Acid Oxidase solution from Crotalus adamanteus, Type IV, minimum 4 units/mg protein (Biuret), Aqueous suspension |
A9378 |
| 1.4.3.2 |
L-Amino Acid Oxidase from Crotalus atrox, Type VI, Dried venom, 0.2-0.3 unit/mg solid |
A5147 |
| 1.4.3.2 |
L-Amino Acid Oxidase from Crotalus adamanteus, BioChemika, powder, yellow, ~0.5 U/mg |
09417 |
| 1.4.3.3 |
D-Amino Acid Oxidase porcine kidney, minimum 0.05 unit/mg solid, Crude powder |
A9128 |
| 1.4.3.3 |
D-Amino Acid Oxidase porcine kidney, Lyophilized powder, minimum 1.5 units/mg protein |
A5222 |
| 1.4.3.3 |
D-Amino Acid Oxidase porcine kidney, BioChemika, suspension, yellow, in 3.6 M ammonium sulfate solution, pH ~6.5, ³15 U/mg protein (~10 mg/ml) |
09412 |
| 1.4.3.3 |
D-Amino Acid Oxidase porcine kidney, BioChemika, powder, yellow, ³0.5 U/mg |
09413 |
| 1.4.3.4 |
Tyramine Oxidase from Arthrobacter sp., approx. 3 units/mg solid |
T0905 |
| 1.4.3.6 |
Diamine Oxidase from porcine kidney, minimum 0.05 unit/mg solid |
D7876 |
| 1.4.3.11 |
L-Glutamate Oxidase from Streptomyces sp., Lyophilized powder, minimum 5 units/mg solid |
G0400 |
| 1.4.3.14 |
Lysine Oxidase from Trichoderma viride, Lyophilized powder, 20-60 units/mg protein |
L6150 |
|
Acting on CH-NH |
|
| 1.5.1.3 |
Dihydrofolate Reductase bovine liver, BioChemika, suspension in 3.6 M ammonium sulfate solution, pH7.0, suspension, off-white, ~8 U/ml |
37294 |
| 1.5.1.5 |
5,10-Methylenetetrahydrofolate Dehydrogenase from Saccharomyces cerevisiae, Type III, suitable for enzymatic assay of tetrahydrofolic acid, Ammonium sulfate suspension, 0.5-1.0 units/mg protein (modified Warburg-Christian) |
M4882 |
| 1.5.1.11 |
Octopine Dehydrogenase from scallops, Lyophilized powder, minimum 5 units/mg protein |
O1252 |
| 1.5.1.7 |
Saccharopine Dehydrogenase, NAD+, lysine-forming from baker's yeast (S. cerevisiae), Lyophilized powder, 40-80 units/mg protein |
S9383 |
| 1.5.3.1 |
Sarcosine Oxidase from Arthrobacter sp., Lyophilized powder, 5-15 units/mg protein |
S8764 |
| 1.5.3.1 |
Sarcosine Oxidase from Bacillus sp., Lyophilized powder, 25-50 units/mg solid |
S7897 |
| 1.5.3.1 |
Sarcosine Oxidase from Corynebacterium sp., Lyophilized powder, 5-10 units/mg protein |
S7759 |
| 1.5.3.x |
Fructosyl-Amino Acid Oxidase from Corynebacterium sp., recombinant, expressed in Escherichia coli, Lyophilized powder, minimum 0.45 units/mg protein |
F7296 |
| 1.5.99.1 |
Sarcosine Dehydrogenase from Pseudomonas sp., Lyophilized powder, 0.5-1.5 units/mg protein |
S4384 |
|
Acting on NAD or NADP |
|
| 1.6.2.4 |
Cytochrome P450 Reductase rabbit liver induced with phenobarbital, approx. 90% (SDS-PAGE), Buffered aqueous glycerol solution, 25-75 units/mg protein (Bradford) |
C4839 |
| 1.6.4.2 |
Glutathione Reductase from spinach, Type VI, Ammonium sulfate suspension, 100-200 units/mg protein |
G3011 |
| 1.6.4.2 |
Glutathione Reductase baker's yeast (S. cerevisiae), Ammonium sulfate suspension, 100-300 units/mg protein (Biuret) |
G3664 |
| 1.6.4.2 |
Glutathione Reductase from wheat germ, Type II, minimum 0.08 units/mg protein |
G6004 |
| 1.6.4.2 |
Glutathione Reductase baker's yeast (S. cerevisiae), BioChemika, in 3.6 M ammonium sulfate solution, pH 7.0, suspension (aqueous), yellow, 100-300 U/mg protein (5-25 mg/ml) |
49755 |
| 1.6.4.2 |
Glutathione Reductase baker's yeast (S. cerevisiae), BioChemika, in 3.6 M ammonium sulfate solution, pH 7.0, suspension (aqueous), yellow, ~100 U/mg protein (~1 mg/ml) |
49756 |
| 1.6.4.5 |
Thioredoxin Reductase from Escherichia coli, >25 units/mg protein (Bradford) |
T7915 |
| 1.6.6.1 |
Nitrate Reductase from corn seedling, minimum 4 units/mL, Buffered aqueous glycerol solution |
N2397 |
| 1.6.6.1 |
Nitrate Reductase from corn seedling, Lyophilized powder, 10-40 units/mg protein |
N9523 |
| 1.6.6.2 |
Nitrate Reductase (NAD[P]H) from Aspergillus niger, Lyophilized powder, minimum 300 units/g solid |
N7265 |
| 1.6.99.2 |
DT Diaphorase from rat, approx. 90% (SDS-PAGE), Lyophilized powder |
D1190 |
| 1.6.99.2 |
DT Diaphorase human, approx. 90% (SDS-PAGE), Lyophilized powder |
D1315 |
| 1.6.99.3 |
NADH-FMN Oxidoreductase from Vibrio harveyi, Lyophilized powder, 0.01-0.05 unit/mg solid |
N3517 |
| 1.6.99.3 |
Cytochrome c reductase from porcine heart, Type I, Lyophilized powder, minimum 1.0 units/mg protein |
C3381 |
| 1.6.99.3 |
NADH-FMN Oxidoreductase from Vibrio harveyi, Lyophilized powder, 0.01-0.05 unit/mg solid |
N3517 |
| 1.6.99.3 |
Cytochrome c reductase from porcine heart, Type I, Lyophilized powder, minimum 1.0 units/mg protein |
C3381 |
| 1.6.99.7 |
Dihydropteridine Reductase from sheep liver, Buffered aqueous glycerol solution, 30-100 units/mg protein (Biuret) |
D6888 |
|
Other nitrogenous donors |
|
| 1.7.3.3 |
Uricase from Candida sp., recombinant, expressed in Escherichia coli, Lyophilized powder, minimum 2 units/mg protein |
U0880 |
| 1.7.3.3 |
Uricase from Arthrobacter globiformis, Lyophilized powder, 15-30 units/mg protein, pH 9.0, 25°C (Biuret) |
U7128 |
| 1.7.3.3 |
Uricase Bacillus fastidiosus, BioChemika, lyophilized, powder, faintly beige, ~15 U/mg |
94310 |
|
Acting on sulfur group donors |
|
| 1.8.1.4 |
Diaphorase Clostridium kluyveri, Lyophilized powder, 5-20 units/mg protein (Biuret, using 2,6-dichlorophenol-indophenol) |
D5540 |
| 1.8.1.4 |
Lipoamide Dehydrogenase from porcine heart, Ammonium sulfate suspension, 2-5 units/mg protein (using 2,6-dichlorophenol-indophenol) |
D3752 |
| 1.8.1.4 |
Lipoamide Dehydrogenase from porcine heart, Type III, Ammonium sulfate suspension, 100-200 units/mg protein (Biuret), Secondary activity: 5-15 units/mg protein Lipoic dehydrogenase, Secondary activity |
L2002 |
| 1.8.1.4 |
Lipoamide Dehydrogenase from bovine intestinal mucosa, Ammonium sulfate suspension, 100-200 units/mg protein (Biuret), Secondary activity: 3-15 units/mg protein Lipoic dehydrogenase, Secondary activity |
L6777 |
| 1.8.1.4 |
Methylthymol Blue sodium salt, indicator for metal titration |
33460 |
| 1.8.1.4 |
Diaphorase Clostridium kluyveri, BioChemika, crystals, light brown-yellow, 6-8 U/mg |
33460 |
| 1.8.3.1 |
Sulfite Oxidase from chicken liver, Ammonium sulfate suspension, 20-70 units/mg protein (Biuret) |
S9526 |
|
Acting on heme group donors |
|
| 1.9.6.1 |
Nitrate reductase (cytochrome) from Escherichia coli, Lyophilized powder, minimum 5 units/g solid |
N0519 |
|
Acting on diphenols and related subs. |
|
| 1.10.3.2 |
Laccase from Rhus vernificera, Crude acetone powder, minimum 50 units/mg solid |
L2157 |
| 1.10.3.2 |
Laccase Coriolus versicolor, BioChemika, powder, light brown, ³0.8 U/mg |
38429 |
| 1.10.3.2 |
Laccase Agaricus bisporus, BioChemika, powder, deep brown, ³4 U/mg |
40452 |
| 1.10.3.3 |
Ascorbate oxidase Cucurbita sp. Lyophilized powder 1,000-3,000 units/mg protein |
A0157 |
| 1.10.3.3 |
Ascorbate oxidase Cucurbita sp., BioChemika, powder, faintly bluish, ³250 U/mg |
11136 |
|
Peroxide as Receptor |
|
| 1.11.1.1 |
NADH Peroxidase from Enterobacter hirae, Ammonium sulfate suspension, minimum 30 units/mg protein (Biuret) |
N6755 |
| 1.11.1.1 |
NADH Peroxidase from Streptococcus faecalis ATCC 11700, recombinant, expressed in Escherichia coli, >95% (SDS-PAGE), Buffered aqueous solution, 50-200 units/mg protein (Biuret) |
N0895 |
| 1.11.1.6 |
Catalase bovine liver, Aqueous solution, minimum 40,000 units/mg protein (E1%/405) |
C3155 |
| 1.11.1.6 |
Catalase bovine liver, Powder, 2,000-5,000 units/mg protein, cell culture tested |
C1345 |
| 1.11.1.6 |
Catalase-Agarose, pH 5.4, Ammonium sulfate suspension, catalase from bovine liver |
C9284 |
| 1.11.1.6 |
Catalase from murine liver, Ammonium sulfate suspension, minimum 900 units/mg protein |
C8531 |
| 1.11.1.6 |
Catalase from human erythrocytes, minimum 90% (SDS-PAGE), Buffered aqueous solution, >30,000 units/mg protein |
C3556 |
| 1.11.1.6 |
Catalase from canine liver, Ammonium sulfate suspension, minimum 900 units/mg protein |
C4138 |
| 1.11.1.6 |
Catalase bovine liver, 2,000-5,000 units/mg protein |
C9322 |
| 1.11.1.6 |
Catalase from bison liver, 2,000-5,000 units/mg protein |
C9447 |
| 1.11.1.6 |
Catalase preparation from Aspergillus niger, Ammonium sulfate suspension, 4,000-8,000 units/mg protein |
C3515 |
| 1.11.1.6 |
Catalase bovine liver, BioChemika, for technical purposes, in 30% glycerol, ³200000 U/ml |
60640 |
| 1.11.1.6 |
Catalase bovine liver, BioChemika, crystalline suspension in water, pH ~6, ~65000 U/mg protein (~20 mg/ml) |
60630 |
| 1.11.1.6 |
Catalase bovine liver, BioChemika, shipped in dry ice, powder, dark green, 10000-25000 U/mg |
60629 |
| 1.11.1.6 |
Catalase bovine liver, BioChemika, lyophilized, powder, brown, ~2500 U/mg |
60635 |
| 1.11.1.6 |
Catalase bovine liver, BioChemika, powder, dark-brown, ~1300 U/mg |
60632 |
| 1.11.1.6 |
Catalase from microorganisms, BioChemika, lyophilized, powder, brown, ~1700 U/mg |
60633 |
| 1.11.1.6 |
Catalase Aspergillus niger, BioChemika, suspension in 3.2 M ammonium sulfate solution, pH6, suspension, dark green-brown, ~70000 U/ml |
60628 |
| 1.11.1.6 |
Catalase Aspergillus niger, BioChemika, ~170 U/mg |
60631 |
| 1.11.1.7 |
Lactoperoxidase bovine milk, Ammonium sulfate suspension, 50-150 units/mg protein |
L2130 |
| 1.11.1.7 |
Peroxidase Acidic Isoenzyme from horseradish, Type X, Ammonium sulfate suspension, approx. 250 units/mg protein (Biuret) |
P6140 |
| 1.11.1.7 |
Peroxidase horseradish, Type II, Essentially salt-free, lyophilized powder, 150-250 units/mg solid |
P8250 |
| 1.11.1.7 |
Peroxidase horseradish, Type I, Essentially salt-free, lyophilized powder, 50-150 units/mg solid |
P8125 |
| 1.11.1.7 |
Peroxidase from Glycine max (soybean), Essentially salt-free, lyophilized powder, 50-150 units/mg solid |
P1432 |
| 1.11.1.7 |
Peroxidase horseradish, Type XII, Essentially salt-free, lyophilized powder, 250-330 units/mg solid |
P8415 |
| 1.11.1.7 |
Peroxidase Highly stabilized from horseradish, Essentially salt-free, lyophilized powder, 200-300 units/mg solid |
P2088 |
| 1.11.1.7 |
Peroxidase horseradish, Type VI, Essentially salt-free, lyophilized powder, 250-330 units/mg solid |
P8375 |
| 1.11.1.7 |
Myeloperoxidase from human leukocytes, Lyophilized powder, minimum 50 units/mg protein |
M6908 |
| 1.11.1.7 |
Peroxidase from Arthromyces ramosus, Lyophilized powder, 50-200 units/mg solid |
P4794 |
| 1.11.1.7 |
Lactoperoxidase bovine milk, Lyophilized powder, 80-120 units/mg protein |
L2005 |
| 1.11.1.7 |
Lactoperoxidase bovine milk, Lyophilized powder, 80-150 units/mg protein |
L8257 |
| 1.11.1.7 |
Peroxidase horseradish, Essentially salt-free, lyophilized powder, 250-330 units/mg solid (using pyrogallol), Type VI-A |
P6782 |
| 1.11.1.7 |
Peroxidase from Arthromyces ramosus, BioChemika, powder, light brown, >400 U/mg |
41366 |
| 1.11.1.7 |
Lactoperoxidase bovine milk, BioChemika, lyophilized, powder, light brown, ~250 U/mg |
61328 |
| 1.11.1.7 |
Lactoperoxidase bovine milk, BioChemika, lyophilized, powder, yellow-brown, ~150 U/mg |
61331 |
| 1.11.1.7 |
Peroxidase from horse radish, BioChemika, lyophilized, powder, red-brown, ~850 U/mg |
77334 |
| 1.11.1.7 |
Peroxidase from horse radish, BioChemika, lyophilized, powder, red-brown, ~700 U/mg |
77333 |
| 1.11.1.7 |
Peroxidase from horse radish, BioChemika, powder, red-brown, ~550 U/mg |
77335 |
| 1.11.1.7 |
Peroxidase from horse radish, BioChemika, lyophilized, powder, beige, ~150 U/mg |
77332 |
| 1.11.1.7 |
Peroxidase from horse radish, BioChemika, essentially salt free, lyophilized, powder, light brown, ~100 U/mg |
77330 |
| 1.11.1.7 |
Myeloperoxidase from human leukocytes, BioChemika, lyophilized, powder, white, ³50 U/mg protein |
70021 |
| 1.11.1.9 |
Glutathione Peroxidase from human erythrocytes, Lyophilized powder, 30-100 units/mg protein |
G4013 |
| 1.11.1.9 |
Glutathione Peroxidase from bovine erythrocytes, Lyophilized powder, 300-700 units/mg protein |
G6137 |
| 1.11.1.9 |
Glutathione Peroxidase from bovine erythrocytes, BioChemika, lyophilized, shipped in dry ice, powder, white, ~100 U/mg |
49753 |
| 1.11.1.10 |
Chloroperoxidase from Caldariomyces fumago, Buffered aqueous suspension, 1,000-2,000 units/mg protein (E1%/280RZ:Approx. 1.0) |
C0887 |
| 1.11.1.10 |
Chloroperoxidase from Caldariomyces fumago, minimum 3,000 units/mL, Buffered aqueous suspension |
C0278 |
| 1.11.1.10 |
Chloroperoxidase from Caldariomyces fumago, BioChemika, suspension in 0.1 M sodium phosphate, pH 4.0, suspension, brown, >10000 U/ml |
25810 |
|
Acting on single donors with incorporation of molecular oxygen |
|
| 1.13.11.12 |
Lipoxidase preparation from Glycine max (soybean), Type I-B, Lyophilized powder, minimum 100,000 units/mg solid |
L7395 |
| 1.13.11.12 |
Lipoxidase preparation from Glycine max (soybean), Type IV, Ammonium sulfate suspension, 300,000-600,000 units/mg protein |
L3004 |
| 1.13.11.12 |
Lipoxidase preparation from Glycine max (soybean), Type V, Ammonium sulfate suspension, 500,000-1,000,000 units/mg protein |
L6632 |
| 1.13.11.12 |
Lipoxidase from Glycine max (soybean), BioChemika, lyophilized, powder, ~9 U/mg |
62340 |
| 1.13.11.3 |
Protocatechuate 3,4-Dioxygenase from Pseudomonas sp., Lyophilized powder, 3-5 units/mg solid |
P8279 |
| 1.13.12.4 |
Lactate Oxidase from Pediococcus sp. |
L0638 |
| 1.13.12.7 |
Lectin, TRITC labeled from Ulex europaeus (gorse, furze), Lyophilized powder |
L4889 |
| 1.13.12.7 |
Luciferase Photinus pyralis (firefly), Lyophilized powder, 10-25 × 106 light units/mg protein |
L9009 |
| 1.13.12.7 |
Luciferase Photinus pyralis (firefly), Lyophilized powder |
L9506 |
| 1.13.12.7 |
Luciferase Photinus pyralis (firefly), Lyophilized powder, minimum 10 × 106 light units/mg protein |
L1759 |
| 1.13.12.7 |
Luciferase-Luciferin reagent in glycine buffer firefly, BioChemika, for ATP-analysis, solid, white |
62646 |
| 1.13.12.7 |
Luciferase Photinus pyralis (firefly), BioChemika, lyophilized, ³85% as protein, powder, white |
62647 |
| 1.13.12.7 |
Luciferase Photinus pyralis (firefly), BioChemika, soluble, ~15% as protein, powder, white |
62648 |
|
Acting on paired donors with incorporation of molecular oxygen |
|
| 1.14.13.- |
2-Tridecanone Monooxygenase from Pseudomonas cepacia, BioChemika, ~0.5 U/g |
91530 |
| 1.14.13.1 |
Salicylate Hydroxylase from Pseudomonas sp., Lyophilized powder, 0.5-3 units/mg protein |
S2907 |
| 1.14.13.16 |
Cyclopentanone Monooxygenase from Pseudomonas sp., BioChemika, powder, off-white, ~10 U/g |
29800 |
| 1.14.13.2 |
p-Hydroxybenzoate Hydroxylase from Pseudomonas sp., Lyophilized powder, approx. 20 units/mg solid |
H9886 |
| 1.14.13.22 |
Cyclohexanone Monooxygenase Acinetobacter sp., BioChemika, powder, off-white, ~5 U/g |
29170 |
| 1.14.13.22 |
Cyclohexanone Monooxygenase Nocardia globerula, BioChemika, powder, grey, ~4 U/g |
29172 |
| 1.14.13.22 |
Cyclohexanone Monooxygenase Xanthobacter sp., BioChemika, powder, off-white, ~6 U/g |
29174 |
| 1.14.13.39 |
Nitric Oxide Synthase, Inducible mouse, Buffered aqueous solution |
N1783 |
| 1.14.13.39 |
Nitric Oxide Synthase, Inducible mouse, recombinant, expressed in Escherichia coli, Buffered aqueous solution |
N2783 |
| 1.14.13.44 |
2-Hydroxybiphenyl 3-Monooxygenase recombinant, BioChemika, solution, slightly yellow, ~2 U/ml |
93043 |
|
Acting on paired donor oxygenases |
|
| 1.14.14.3 |
Luciferase from Vibrio fischeri (Photobacterium f), Lyophilized powder |
L8507 |
| 1.14.14.3 |
Luciferase from Vibrio fischeri (Photobacterium f), BioChemika, lyophilized, ~8% as protein, powder |
62641 |
| 1.14.15.1 |
(+)-Camphor Monooxygenase from Pseudomonas putida, BioChemika, ~0.3 U/g |
21332 |
| 1.14.18.1 |
Tyrosinase mushroom, ³2,000 units/mg solid |
T7755 |
| 1.14.18.1 |
Tyrosinase mushroom, BioChemika, lyophilized, powder, brown, ³2000 U/mg |
93898 |
| 1.14.99.4 |
Progesterone Monooxygenase from Cylindrocapron radicicola, BioChemika, powder, beige, ~0.6 U/g |
81703 |
|
Acting on superoxide radicals |
|
| 1.15.1.1 |
Superoxide Dismutase from Spirulina platensis, minimum 10% (remainder potassium phosphate), Lyophilized powder, minimum 500 units/mg protein |
S3813 |
| 1.15.1.1 |
Superoxide Dismutase bovine erythrocytes, cell culture tested, Lyophilized powder, 2,500-7,000 units/mg protein |
S5395 |
| 1.15.1.1 |
Superoxide Dismutase from horseradish, Lyophilized powder, 1,000-4,000 units/mg protein |
S4636 |
| 1.15.1.1 |
Superoxide Dismutase from Escherichia coli, Lyophilized powder, 2,500-5,000 units/mg protein |
S5639 |
| 1.15.1.1 |
Superoxide Dismutase from Escherichia coli, Lyophilized powder, minimum 2,500 units/mg protein |
S5389 |
| 1.15.1.1 |
Superoxide Dismutase canine erythrocytes, Lyophilized powder, 2,000-6,000 units/mg protein |
S9511 |
| 1.15.1.1 |
Superoxide Dismutase bovine liver, Ammonium sulfate suspension, approx. 3,000 units/mg protein (Biuret) |
S7008 |
| 1.15.1.1 |
Superoxide Dismutase bovine erythrocytes, Lyophilized powder, 2,500-7,000 units/mg protein |
S2515 |
| 1.15.1.1 |
Superoxide Dismutase bovine liver, Lyophilized powder, 2,000-6,000 units/mg protein |
S8160 |
| 1.15.1.1 |
Superoxide Dismutase from human erythrocytes, Lyophilized powder, 3,000-6,000 units/mg protein |
S9636 |
| 1.15.1.1 |
Superoxide Dismutase bovine erythrocytes, BioChemika, powder, blue-grey, 2'000-10'000 U/mg |
86200 |
|
Acting on ferredoxin |
|
| 1.18.1.2 |
Ferredoxin-NADP+ Reductase from Spinacia oleracea (spinach), Lyophilized powder, 2-6 units/mg solid, Secondary activity: approx. 15 NADPH diaphorase |
F0628 |
| |
|
back to top |
Oxidoreductases | Transferases | Hydrolases (Acting on esters) | Hydrolases (Glycosidases) | Hydrolases (Other) | Lyases | Isomerases | Ligases