|
Specifications
|
Activity:
|
3–8 TIU/mg solid
|
|
Purity:
|
Minimum 98% by SDS–PAGE
|
|
Unit Definition:
|
One Trypsin Inhibitor Unit (TIU) will decrease the activity of 2 trypsin units by 50%, where 1 trypsin unit will hydrolyze 1.0 µmole of N-α-benzoyl-DL-arginine p-nitroanilide (BAPNA) per minute at pH 7.8 and 25°C. Another commonly used unit of activity is the KIU (Kallikrein Inhibitor Unit). From our data, a conversion factor for Aprotinin is: 1 TIU equals ~1,300 KIU.
|
|
Appearance:
|
White to off-white powder
|
|
Cell Culture Test:
|
Pass
|
3 lots with Certificates of Analysis available. For Bulk Quantities, please call 1-800-336-9179.
|
Product Description
Aprotinin is a competitive serine protease inhibitor which forms stable complexes with and blocks the active sites of enzymes. The binding is reversible, and most aprotinin-protease complexes dissociate at pH >10 or <3.2. Molecular Weight: ~ 6511 E1%(280 nm) = 8.3 (water)
Stability / Storage as Supplied
If stored at 2-8°C Product Code A6103 has a designated shelf-life of two years.
|
|
Solubility / Solution Stability
Aprotinin is freely soluble in water (>10 mg/mL) and in aqueous buffers of low ionic strengths. Dilute solutions are generally less stable than concentrated ones. Solution stability also depends on pH; values of 1-12 can be tolerated. Repeated freeze-thaw cycles should be avoided. The Cys14-Cys38 disulfide bridge is readily split by reducing agents like 2-Mercaptoethanol. Due to its compact tertiary structure, aprotinin is relatively stable against denaturation due to high temperature, acids, alkalies, organic solvents or proteolytic degradation (only thermolysin has been found capable of degrading aprotinin after heating to 60-80°C). The high basicity of aprotinin causes it to adhere to commonly used dialysis tubing and even gel filtration matrices, but the use of acetylated materials and concentrated salt solutions (e.g., 0.1 M NaCl in buffer)3 minimizes the problem. Sterilization may be achieved by filtration through a 0.2 µm filter.
| Enzyme |
Inhibition |
| Acrosin |
Weak inhibition6 |
| Chymotrypsin |
Ki = 9 nM9 |
| Chymotrypsinogen (bovine), pH 8.0 |
Ki = 9 nM4 |
| CMP-Sialic Acid: Lactosylceramide -(2,3)-Sialyltransferase |
74% Inhibition at 300 nM9 |
| Elastase (human leukocytes), pH 8.0 |
Ki = 3.5 µM4 |
| Kallikrein (pancreatic), pH 8.0 |
Ki = 1.0 nM4 |
| Kallikrein (plasma) |
Ki = 30 nM; 100 nM9 |
| Kallikrein (tissue) |
Ki = 1 nM9 |
| Kallikrein (urine) |
Ki = 1.7 nM9 |
| Plasmin (porcine), pH 7.8 |
Ki = 4.0 nM4 |
| Plasminogen activator |
Ki = 8 µM; 27 µM9 |
| Trypsin (bovine), pH 8.0 |
Ki = 0.06 pM4 |
| Trypsinogen (bovine), pH 8.0 |
Ki = 1.8 µM4 |
| Tryptase TL-2 |
16% Inhibition at 10 µM9 |
| Urokinase (human), pH 8.8 |
Ki = 8.0 µM4 |
References
- Merck Index, 12th Ed., S. Budavari, Ed., # 796, p. 128 (1996).
- J. Gen. Physiol., 19, 991 (1936).
- Hoppe-Seyler's Z. Physiol. Chem., 192,1 (1930).
- Drug Res., 33(1), No. 4, 479 (1983).
- Sigma data.
- Biochemica Information, 1st Ed., J. Keesey, Ed., Boehringer Mannheim Biochemicals, p. 111, Indianapolis (1987).
- Biochem., 7, 3634 (1968).
- Life Sci., 28, 1861 (1981).
- Handbook of Enzyme Inhibitors, 2nd Ed., Part B, H. Zollner, Ed., p. 572, VCH Verlagesgesellschaft, Weinheim (1993).