Sigma-Aldrich
Life Science > Metabolomics > Enzyme Explorer > Enzyme Reagents > Protease Detection Kits
Enzyme Explorer
Protease Detection Kits
Now there are two easy-to-use kits to detect trace amounts of protease or determine total protease activity using the same methodologies utilized by Sigma's QC department for years. Sigma has used these protocols to test thousands of samples for proteolytic activity.
The Protease Detection Kits are complete kits for detection of primary or trace protease activity by your choice of fluorometric or colorimetric detection. Everything you need, including control, standards, buffers, and substrate, is included. For convenience the assay can be performed in either a cuvette or microplate format. Each kit contains enough reagents for 200 x 1 mL assays. For greater sensitivity when measuring low levels of protease activity, the Protease Fluorescent Detection kit (PF0100) is recommended. For more accurate quantification of primary protease activity, the Protease Colorimetric Detection kit is (PC0100) recommended.
Both the Fluorometric Detection Kit and the Colorimetric Detection Kit have been tested against representative samples of all four types of protease classes (serine, aspartic, cysteine and metalloproteases) to ensure broad application suitability.
Protease Detection Kits provide the following benefits:
  • Complete kits include controls, standards, buffers and substrate
    Saves time and eliminates variables.
  • Detects wide range of proteases (serine, aspartic, cysteine and metalloproteases)
    Gives you the confidence you need to detect proteases of unknown specificity.
  • Simple and easy to use with a comprehensive technical bulletin
    Save your research efforts for discovery and not routine analysis.
  • Can be used with either cuvette or microplate formats
    Analyze a single sample or use for high throughput
  • Contains positive control with known specific activity
    Guaranteed reliability
Protease Fluorescent Detection Kit, PF0100
The Protease Fluorescent Detection Kit is based on an assay method using highly labeled fluorescein isothiocyanate (FITC) casein as the substrate. This is a modified version of the procedure for protease detection as described by Twining.[1] The kit will detect 0.5 µg/mL of protease (approx. 5 ng trypsin) and has been tested for detection of all four proteolytic classes (serine, aspartic, cysteine and metalloproteases). The sensitivity level can be increased by using longer incubation times of up to 24 hours.
PFD_Kit
Detection limits for the Protease Fluorescent Detection Kit
Protease Protease Class Detection Limits Cleavage Site(s)1
Trypsin Serine < 1.0 ng C-terminal sites of Arg and Lys
Pepsin Aspartic 1.5 ng Hydrophobic, preferably aromatic, residues in P1 and P1' positions.
Papain Cystine 1.5 ng Hydrolysis of proteins with broad specificity for peptide bonds, but preference for an amino acid bearing a large hydrophobic side chain at the P2 position. Does not accept Val in P1'
Carboxypeptidase A Metalloproteinase 5.0 ng Release of a C-terminal amino acid, but little or no action with -Asp, -Glu, -Arg, -Lys or -Pro
Data Points
sensitivity-of-pf001   typical-activity-pf001
Sensitivity of the Protease Fluorescent Detection Kit (PF0100) using Trypsin (T6567)
Each of the trypsin samples was incubated with the proprietary formulation of the FITC-casein substrate for 17 hours for increased sensitivity at 37°C and measured at an excitation wavelength of 485 nm and an emission wavelength of 535 nm using a microtiter plate.
  Typical activity curve using trypsin
Each of the trypsin samples was incubated with the proprietary formulation of the FITC-casein substrate for 30 minutes at 37°C and measured at an excitation wavelength of 485 nm and an emission wavelength of 535 nm using a microtiter plate.
Kit Components
  • Incubation Buffer
  • Assay Buffer
  • FITC-casein substrate
  • 0.6 N TCA solution
  • Trypsin, protease control
  • Fluorescein isothiocyanate (FITC) control
Ordering Information
Product No. Product Name Package Size Technical Manual
PF0100 Protease Fluorescent Detection Kit 1 KT sg_dd_hts_tbicon
Reference
  1. Twining, S.S., Fluorescein Isothiocyanate-Labeled Casein Assay for Proteolytic Enzymes. 1984. Anal. Biochem., 143, 30-34 (1984).
Protease Colorimetric Detection Kit, PC0100
The Protease Colorimetric Detection Kit provides robust, linear response with convenient colorimetric detection.
The kit is based on the cleavage of casein into trichloroacetic acid-soluble peptides containing tyrosine and tryptophan residues. These peptides react with Folin-Ciocalteu reagent to give a characteristic blue color that can be measured at 660 nm. The kit will detect approximately 0.1 units/ml of protease (approx. 2 µg), and has been tested for detection of all four proteolytic classes (serine, aspartic, cysteine and metalloproteases).
PCD_Kit
Kit Components
  • Protease Detection Substrate: Casein
  • Folin & Ciocalteu’s phenol reagent (Product No. F9252)
  • 500 mM Sodium carbonate solution
  • Enzyme diluent
  • 0.6 N TCA
  • Tyrosine standard solution
  • Protease from Bacillus polymyxa control
Ordering Information
Product No. Product Name Package Size Technical Manual
PC0100 Protease Colorimetric Detection Kit 1 KT sg_dd_hts_tbicon
back to top
back to Enzyme Explorer