Enzyme Explorer
Molecular Parameters
|
Prealbumin, thyroxine-binding Retinol-binding protein Albumin α-Globulins β-Globulins Apolipoproteins γ-Mobility (non-immunoglobulin) Complement Components Coagulation Proteins Cell-Related Plasma Proteins Immunoglobulins (Antibodies) Amyloid Proteins (Non-Immunoglobulin) Misc. Trace Components
Plasma & Blood Protein Resource
| Prealbumin, thyroxine-binding |
| Protein |
Species |
Subunit Structure |
Method |
Chain Length |
Protein MW |
Amino Terminus |
Carb. Content (%) |
| Prealbumin, thyroxine-bindin |
Human |
Noncovalent tetramer of identical chains |
AA |
127 |
54,980 |
Gly |
0 |
| Retinol-binding protein |
back to top |
| Protein |
Species |
Subunit Structure |
Method |
Chain Length |
Protein MW |
Amino Terminus |
Carb. Content (%) |
| Retinol-binding protei |
Human |
Single chain |
AA |
182 |
20,957 |
Glu |
0 |
| Albumin |
back to top |
| Protein |
Species |
Subunit Structure |
Method |
Chain Length |
Protein MW |
Amino Terminus |
Carb. Content (%) |
| Albumin |
Human |
Single chain |
AA |
585 |
66,458 |
Asp |
0 |
| |
|
Nucleic acid precursor |
NA |
609 |
69,365 |
|
0 |
| |
Bovine |
Single chain |
AA |
582 |
66,210 |
Asp |
0 |
| |
|
Nucleic acid precursor |
NA |
606 |
|
|
|
| |
Porcine |
Single chain |
AA |
~575 |
~66,300 |
Asp |
0 |
| |
Ovine |
Single chain |
AA |
575 |
~66,300 |
Asp |
0 |
| |
Chicken |
Single chain |
AA |
575 |
~66,300 |
Asp |
0 |
| |
Rat |
Single chain |
AA |
584 |
64,600 |
Glu |
0 |
| |
|
Nucleic acid precursor |
NA |
608 |
|
Glu |
|
| α-Globulins |
back to top |
| Protein |
Species |
Subunit Structure |
Method |
Chain Length |
Protein MW |
Amino Terminus |
Carb. Content (%) |
| α-Globulins |
|
| α1-Microglobulin |
Human |
Single chain |
AA |
167 |
26,000 |
Gly |
20 |
| α1-Acid glycoprotein |
Human |
Single chain |
AA |
181 |
40,000 |
pGlu |
42 |
| |
Rat |
Nucleic acid precursor |
NA |
187 |
~40,000 |
Gln |
+ |
| α1-Antitrypsin |
Human |
Single chain |
AA |
394 |
54,000 |
Glu |
13 |
| |
|
Nucleic acid precursor |
NA |
|
| |
Baboon |
Nucleic acid precursor |
NA |
394 |
~54,000 |
Glu |
+ |
| α1-Antichymortrypsin |
Human |
Single chain |
AA |
~440 |
68,000 |
Asn |
26 |
| |
|
|
NA |
408 |
|
|
|
| α2-Antiplasmin |
Human |
Single chain |
AA |
|
65,000 |
Asn |
11 |
| α1-Fetoprotein |
Human |
Nucleic acid precursor |
NA |
590 |
66,300 |
Thr |
3 |
| |
Rat |
Nucleic acid precursor |
NA |
|
| |
Mouse |
Nucleic acid precursor |
NA |
586 |
72,000 |
Ala |
|
9.5 S α1-Glycoprotein (serum amyloid P component) |
Human |
Two pentagons (10 non-covalently bound identical subunits) |
AA |
(182)10 |
~250,000 |
His |
13 |
| α1B-Glycoprotein |
Human |
Possibly a single chain |
AA |
~480 |
68,000 |
Ala |
13 |
| α1T-Glycoprotein |
Human |
Single chain |
|
~776 |
85,000 |
|
15 |
Vitamin D-binding protein (Gc globulin) |
Human |
Single chain |
AA |
~450 |
52,000 |
Leu |
4 |
| Thyroxine-binding protein |
Human |
Single chain |
|
|
54,000 |
|
20 |
| Inter-α-trypsin inhibitor |
Human |
Single chain |
AA |
|
~160,000 |
Lys |
8 |
| Ceruloplasmin |
Human |
Single chain (easily cleaved) |
AA |
1,046 |
132,000 |
Lys |
7 |
| 3.8 S Histidine-rich α2-glycoprotein |
Human |
Single chain |
AA |
~440 |
~58,500 |
Val |
11 |
| Galactoglycoprotein |
Human |
Single chain |
AA |
~213 |
81,000 |
Blocked |
76 |
| 3.1 S Leucine-rich α2-glycoprotein |
Human |
Single chaine |
AA |
312 |
50,000 |
Val |
23 |
| α2µ-Globulin |
Rat (male) |
Nucleic acid precursor |
NA |
162 |
18,709 |
Glu |
+ |
| Zn-α2-Glycoprotein |
Human |
Single chain |
|
~258 |
41,000 |
|
18 |
| α2HS-Glycoprotein |
Human |
Two chains (A & B) |
AA |
~359 |
49,000 |
Ala |
14 |
| |
|
B chain |
AA |
27 |
3,386 |
Thr |
+ |
| α2-macroglobulin |
Human |
4 identical subunits |
AA |
1,450 |
725,000 |
Ser |
9.4 |
| 4S-α2,β1-Glycoprotein |
Human |
Single chain |
|
~465 |
60,000 |
Arg |
28 |
| Haptoglobin |
Human |
2 pairs of nonidentical chains (αβ)2 |
AA |
656 |
86,018 |
Val/Ile |
16.4 |
| |
|
Single-chain precursor |
NA |
245 |
|
|
|
| |
|
α1 chain |
AA |
83 |
9,189 |
Val |
0 |
| |
|
α2 chain |
AA |
142 |
15,939 |
Val |
0 |
| |
|
β chain |
AA |
245 |
33,820 |
Ile |
19.4 |
| |
|
β chain |
NA |
|
| |
Canine |
One pair of chains (αβ) β chain |
AA |
|
| Corticosteroid-binding globulin |
Human |
Single chain |
|
|
52,000 |
|
30 |
| 8 S α3-Glycoprotein |
Human |
Tetramer |
|
|
220,000 |
|
31 |
| β-Globulins |
back to top |
| Protein |
Species |
Subunit Structure |
Method |
Chain Length |
Protein MW |
Amino Terminus |
Carb. Content (%) |
| Pregnancy-specific β1-glycoprotein Human |
A trimeric aggregate |
|
|
(42,300)n |
|
32.6 |
| Transferrin |
Human |
Single chain |
AA |
678 |
79,550 |
Val |
5.9 |
| |
|
|
NA |
679 |
|
Val |
|
| Ovotransferrin |
Chicken |
Single chain |
AA |
686 |
77,770 |
Ala |
+ |
| |
|
Nucleic acid precursor |
NA |
686 |
|
|
|
| Hemopexin |
Human |
Single chain |
AA |
439 |
60,000 |
Thr |
22 |
| |
Rabbit |
Single chain |
AA |
~440 |
57,000 |
Val |
20 |
| β2-Microglobulin |
Human |
Single chain |
AA |
99 |
11,731 |
Ile |
0 |
| |
Mouse |
Single chain |
AA |
99 |
|
Ile |
0 |
| |
Rabbit |
Single chain |
AA |
99 |
|
Val |
0 |
| |
Guinea pig |
Single chain |
AA |
99 |
|
Val |
0 |
| C-Reactive protein |
Human |
Subunits aggregate as cyclic pentamers |
AA |
187 |
(20,946)5 |
pGlu |
0 |
| |
Rabbit |
Pentagonal structure |
AA |
186 |
|
Ala |
0 |
| β2-Glycoprotein I |
Human |
Single chain |
AA |
326 |
~50,000 |
Gly |
19 |
| β2-Glycoprotein III |
Human |
Single chain, forms complex |
AA |
|
35,000 |
|
10 |
| Apolipoproteins |
back to top |
| Protein |
Species |
Subunit Structure |
Method |
Chain Length |
Protein MW |
Amino Terminus |
Carb. Content (%) |
| Apolipoprotein A-I (HDL) |
Human |
Single chain; no disulfides |
AA |
243 |
28,076 |
Asp |
0 |
| Apolipoprotein A-II (HDL) |
Human |
Dimer of 2 identical chains, joined by disulfide |
AA |
77 |
17,414 |
pGlu |
0 |
| Apolipoprotein C-I (VLDL) |
Human |
Single chain; no disulfides |
AA |
57 |
6,631 |
Thr |
0 |
| Apolipoprotein C-II |
Human |
Single chain; no disulfides |
AA |
78 |
8,829 |
Thr |
0 |
| Apolipoprotein C-III (VLDL) |
Human |
Single chain glycopeptide |
AA |
79 |
8,764 |
Ser |
3 |
| Apolipoprotein E |
Human |
Single chain |
AA |
299 |
34,183 |
Lys |
+ |
| γ-Mobility (non-immunoglobulin) |
back to top |
| Protein |
Species |
Subunit Structure |
Method |
Chain Length |
Protein MW |
Amino Terminus |
Carb. Content (%) |
| 0.6 S γ2-Globulin |
Human |
Single chain |
AA |
47 |
5,100 |
Phe |
0 |
| 2 S γ2-Globulin |
Human |
Single chain |
AA |
132 |
14,000 |
Tyr |
0 |
| Basic Protein B2 |
Human |
Single chain |
AA |
~85 |
9,000 |
|
0 |
| Post-γ-Globulin (γ-trace) |
Human |
Single chain |
AA |
120 |
13,248 |
Ser |
0 |
| Complement Components |
back to top |
| Protein |
Species |
Subunit Structure |
Method |
Chain Length |
Protein MW |
Amino Terminus |
Carb. Content (%) |
| C1q |
Human |
Six identical nocovalently-linked subunits, each with three similar chains (A,B&C) Total of 18 chains |
AA |
|
410,000 |
Glu/Asn |
9 |
| |
|
B chain |
AA |
226 |
|
Glu |
+ |
| |
|
C chain |
AA |
~200 |
|
Asn |
+ |
| C1r |
Human |
Noncovalent dimer |
|
|
(83,000)2 |
|
9.4 |
| C1r |
Human |
B chain |
AA |
242 |
27,096 |
Ile |
12.4 |
| C1s |
Human |
Single chain |
AA |
|
83,000 |
Glu |
7.1 |
| C1s |
Human |
Two chains (a and b) |
AA |
|
83,000 |
Glu/Ile |
7.1 |
| C2 |
Human |
Single chain |
AA |
|
102,000 |
Ala |
+ |
| C3 |
Human |
Two disulfide-bonded chains |
AA |
~1,630 |
~185,000 |
|
2 |
| |
|
α chain |
AA |
~960 |
115,000 |
Ser |
|
| |
|
β chain |
AA |
~670 |
75,000 |
Ser |
|
| |
Mouse |
ProcC3 is a single-chain precursor |
NA |
|
~190,000 |
Ile |
|
| C3a |
Human |
Fragment of C3 a chain |
AA |
77 |
9,093 |
Ser |
0 |
| |
Porcine |
Fragment of C3 α chain |
AA |
77 |
~9,000 |
Ser |
|
| |
Rat |
Fragment of C3 α chain |
AA |
78 |
~9,000 |
Ser |
|
| |
Mouse |
Fragment of C3 α chain |
NA |
78 |
~9,800 |
Ser |
|
| C4 |
Human |
Three disulfide-bonded chains |
AA |
|
|
7 |
|
| |
|
α chain |
AA |
~785 |
93,000 |
|
|
| |
|
β chain |
AA |
~660 |
75,000 |
|
|
| |
|
γ chain |
AA |
~250 |
33,000 |
|
|
| C4a |
Human |
Fragment of a chain |
AA |
77 |
8,763 |
Asn |
0 |
| |
Bovine |
Fragment of a chain |
AA |
77 |
|
|
|
| C5 |
Human |
Two disulfide-bonded chains |
|
|
185,000 |
|
1.6 |
| |
|
α chain |
AA |
~960 |
115,000 |
Thr |
|
| |
|
β chain |
|
~670 |
75,000 |
|
|
| C5a |
Human |
Fragment of α chain |
AA |
74 |
8,274 |
Thr |
+ |
| C6 |
Human |
Single chain |
AA |
~900 |
1,048,000 |
Ala |
3.8 |
| C7 |
Human |
Single chain |
AA |
~800 |
92,400 |
Ser |
6.4 |
| C8 |
Human |
Two noncovalently-linked subunits |
|
|
163,000 |
|
|
| C4-Binding Protein |
Human |
Disulfide-bonded subunits (Mr 75,000) |
|
|
540,000 |
Asn |
|
| Factor D (C3 Proactivator Convertase) |
Human |
Single chain |
AA |
|
24,000 |
Ile |
+ |
| Factor B (C3 Proactivator) |
Human |
Single chain |
AA |
|
90,000 |
Thr |
6 |
| |
|
Bb Fragment |
AA |
505 |
60,000 |
Lys |
+ |
| Properdin |
Human |
Four non-covalent identical chains |
AA |
~480 |
220,000 |
Asp |
10 |
| Factor H (β1H) |
Human |
Single chain |
AA |
~1,275 |
155,000 |
Glu |
9.3 |
| C3b-Inactivator |
Human |
Two disulfide-bonded chains |
|
|
90,000 |
Ile/Lys |
11 |
| |
|
Light chain |
AA |
|
40,000 |
Ile/Lys |
+ |
| |
|
Heavy chain |
AA |
|
50,000 |
Lys |
+ |
| Coagulation Proteins |
back to top |
| Protein |
Species |
Subunit Structure |
Method |
Chain Length |
Protein MW |
Amino Terminus |
Carb. Content (%) |
| Prothrombin |
Human |
Single chain |
AA |
581 |
72,000 |
Ala |
12 |
| |
Bovine |
Single chain |
AA |
582 |
72,000 |
Ala |
12 |
| Thrombin |
Bovine |
Two disulfide-linked chains |
AA |
~340 |
37,000 |
Thre/ILE |
|
| |
|
A chain |
AA |
49 |
5,728 |
Thre/ILE |
0 |
| |
|
B chain |
AA |
259 |
31,000 |
Ile |
|
| FactorVII |
Human |
Single chain (two chains when activated) |
AA |
|
48,000 |
Ala |
16 |
| Factor VIII |
Human |
Noncovalent complex |
|
|
(100,000)n |
|
6 |
| Factor IX |
Human |
Single chain Precursor |
NA |
462 |
51,801 |
|
+ |
| |
Bovine |
Single chain |
AA |
427 |
~55,000 |
Tyr |
26 |
| Factor IXa (activated) |
Human |
Heavy and light chains |
NA |
381 |
|
|
|
| |
|
Light chain |
NA |
145 |
|
Tyr |
|
| |
|
Heavy chain |
AA |
236 |
|
Val |
|
| |
Bovine |
Heavy and light chains |
AA |
390 |
~46,500 |
Tyr/Val |
11 |
| |
|
Light chain |
AA |
146 |
16,600 |
Tyr |
7.4 |
| |
|
Heavy chain |
AA |
232 |
27,300 |
Val |
13 |
| Factor X |
Human |
Heavy and light chains |
AA |
|
59,000 |
Ala/Ser |
15 |
| |
|
Light chain |
AA |
139 |
16,211 |
Ala/Ser |
0 |
| |
Bovine |
Heavy and light chains |
AA |
447 |
55,000 |
Trp/Ala |
10 |
| |
|
Light chain |
AA |
140 |
16,143 |
Ala/Ser |
0 |
| |
|
Heavy chain |
AA |
307 |
38,000 |
Trp/Ala |
11.5 |
| Factor Xa Activated |
Bovine |
Heavy and light chains |
AA |
396 |
44,000 |
Ile/Ala |
3 |
| |
|
Light chain |
AA |
140 |
16,000 |
Ala |
0 |
| |
|
Heavy chain |
AA |
256 |
28,000 |
Ile |
0 |
| Factor XI |
Human |
Two idenical disulfide-linked chains |
AA |
|
130,000 |
Gly |
5 |
| Protein C |
Bovine |
Two disulfide-linked chains |
AA |
415 |
55,000 |
Ala/Asp |
14 |
| |
|
Light chain |
AA |
155 |
21,000 |
Ala |
+ |
| |
|
Heavy chain |
AA |
260 |
35,000 |
Asp |
+ |
| Protein S |
Human |
Single chain |
AA |
|
~69,000 |
Ala |
7.8 |
| |
Bovine |
|
AA |
|
~66,000 |
Ala |
+ |
| Protein Z |
Bovine |
Single chain |
AA |
|
~50,000 |
Ala |
+ |
| Fibrinogen |
Human |
Three pairs of disulfide bonded chains |
AA |
2,964 |
340,000 |
|
4 |
| |
|
α chain |
AA |
610 |
64,115 |
Ala |
|
| |
|
β chain |
AA |
461 |
52,314 |
Gln |
+ |
| |
|
γ chain |
AA |
411 |
46,468 |
Tyr |
+ |
| Plasminogen |
Human |
Single chain |
AA |
790 |
~92,000 |
Glu |
1.5 |
| Plasmin |
|
Two-chain after activation |
|
790 |
|
Glu/Val |
1.5 |
| |
|
Heavy chain (A) |
|
560 |
|
Glu |
+ |
| |
|
Light chain (B) |
|
230 |
|
Val |
0 |
| Factor XII |
Human |
Single chain |
AA |
|
76,000 |
Ile |
17 |
| Factor XIII (Transglutaminase) |
|
Two pairs of non-identical chains (a2b2) |
AA |
|
320,000 |
|
4.9 |
| HMW and LMW Prekininogens |
Bovine |
Single chain Precursor |
NA |
621 |
|
|
|
| LMW Kininogen |
Bovine |
Heavy chain |
AA |
361 |
|
(Thr) |
+ |
| |
|
Light chain |
|
47 |
|
Ser |
|
| |
|
| Antithrombin III |
Human |
Single chain |
AA |
430 |
58,000 |
His |
15 |
| Cell-Related Plasma Proteins |
back to top |
| Protein |
Species |
Subunit Structure |
Method |
Chain Length |
Protein MW |
Amino Terminus |
Carb. Content (%) |
| Fibronectin |
Human |
Disulfide-linked Dimer |
AA |
~1,900 |
~440,000 |
pGlu |
+ |
| |
Human |
Disulfide-linked Dimer |
AA |
~1,880 |
~440,000 |
pGlu |
+ |
| β-Thromboglobulin |
Human |
Tetramer of identical chains |
AA |
81 |
35,404 |
Gly |
0 |
| Platelet Factor-4 |
Human |
Tetramer of identical chains |
AA |
70 |
(7,769)4 |
Glu |
0 |
| Serum Basic Protease Inhibitor |
Bovine |
Single chain |
AA |
60 |
6,647 |
Thr |
0 |
| Immunoglobulins (Antibodies) |
back to top |
| Protein |
Species |
Subunit Structure |
Method |
Chain Length |
Protein MW |
Amino Terminus |
Carb. Content (%) |
| Immunoglobulins (Antibodies) |
Higher Vertebrates |
Four disulfide-bonded chains, two heavy and two light |
|
|
(160,000)n |
|
Varies |
| κ or λ Light Chains (Bence Jones Proteins) |
Human |
Monomer of dimer |
AA |
~214 |
~23,000 |
Varies |
0 |
| |
Human |
Kappa (κ) |
AA |
~214 |
~23,500 |
Varies |
0 |
| |
|
Lambda (λ) |
AA |
~214 |
~23,000 |
Varies |
0 |
| Immunoglobulin G (IgG) |
Human |
(γκ)2 or (γλ)2 monomer |
|
|
~ 150,000 |
Varies |
3 |
| IgG1 |
|
γ1 Heavy chain |
AA |
~475 |
~50,000 |
Varies |
+ |
| IgG2 |
|
γ2 Heavy chain |
AA |
~450 |
~50,000 |
Varies |
+ |
| IgG3 |
|
γ3 Heavy chain |
AA |
~475 |
~55,000 |
Varies |
+ |
| IgG4 |
|
γ4 Heavy chain |
AA |
~450 |
~50,000 |
Varies |
+ |
| Immunoglobulin A (IgA) |
Human |
(α2κ2)n or (α2λ2)n n=1,2 or 4 |
|
|
~(160,000)n |
Varies |
8 |
| IgA1 |
|
α1 Heavy chain |
AA |
~475 |
~60,000 |
Varies |
+ |
| IgA2 |
|
α2 Heavy chain |
AA |
~450 |
~60,000 |
Varies |
+ |
| Immunoglobulin M (IgM) |
Human |
µ2κ2 or µ2λ2 monomer |
|
|
~950,000 |
Varies |
10 |
| |
|
µ Heavy chain |
AA |
~575 |
~75,000 |
Varies |
+ |
| Immunoglobulin D (IgE) |
Human |
δ2κ2 or δ2λ2 monomer |
|
~510 |
~175,000 |
Varies |
9 |
| |
|
δ Heavy chain |
AA |
|
~65,000 |
Varies |
+ |
| Immunoglobulin E (IgE) |
Human |
σ2κ2 or σ2λ2 monomer |
|
|
~190,000 |
Varies |
12 |
| |
|
σ Heavy chain |
AA |
~550 |
~72,000 |
Varies |
+ |
| J Chain |
Human |
Single chain |
AA |
129 |
14,619 |
pGlu |
|
| |
|
Single chain Precursor |
NA |
137 |
|
|
|
| Amyloid Proteins (Non-Immunoglobulin) |
back to top |
| Protein |
Species |
Subunit Structure |
Method |
Chain Length |
Protein MW |
Amino Terminus |
Carb. Content (%) |
| Amyloid-Related Apoprotein (apoSAA1) |
Human |
Single chain Precursor |
AA |
104 |
11,683 |
Arg |
0 |
| AA (FMF) (ASF) |
Human |
Single chain Precursor |
AA |
76 |
9,150 |
Arg |
0 |
| AA (TH) (AS) |
Human |
Single chain Precursor |
AA |
76 |
9,145 |
Arg |
0 |
| Serum Amyloid P Component (9.5 S 7α1-glycoprotein) |
Human |
Ten Subunits aggregates as two pentamers |
AA |
~184 |
~25,000 |
His |
13 |
| Misc. Trace Components |
back to top |
| Protein |
Species |
Subunit Structure |
Method |
Chain Length |
Protein MW |
Amino Terminus |
Carb. Content (%) |
| Carcinoembryonic Antigen |
Human |
AA |
|
+ |
| Angiotensinogen |
Human |
AA |
|
|
|
Asp |
|
| |
Rat |
Single chain Precursor |
NA |
453 |
49,548 |
Asp |
(+) |
back to top
A special thanks to Frank Putnam for allowing us to replicate information referenced from "The Plasma Proteins", Volume IV, 2nd Edition, 1984, Frank W. Putnam.