Enzyme Explorer
Broad Spectrum Inhibitors
|
Serine Protease Inhibitors Cysteine Protease Inhibitors Aspartic Protease Inhibitors Metalloproteinase Inhibitors References
Serine Protease Inhibitors
| Product Code |
Inhibitor |
Characteristics |
Typical Working Concentration |
Prep. of Stock Solutions |
| L2884 |
Leupeptin |
Inhibits trypsin-like serine proteases such as trypsin, chymotrypsin, chymase, pepsin and thrombin. Inhibits selected cysteine proteases such as calpain, cathepsin B, H & L and papain. |
10-100 µM |
10 mM in water, stable 6 months at -20 °C |
| P7626 |
PMSF |
Broad spectrum serine protease inhibitor. Also reported to inhibit some cysteine proteases such as papain. |
0.1-1.0 mM |
Prepare fresh in anhydrous ethanol or isopropanol at 200 mM |
| A8456 |
AEBSF |
Broad spectrum serine protease inhibitor. Also reported to inhibit some cysteine proteases such as papain. |
0.1-1.0 mM |
100 mM in water, stable 1 month at -20 °C |
| A1153 |
Aprotinin |
Does not inhibit thrombin or factor Xa. |
0.3 µM or equimolar |
Freely soluble in water and stable at 2-8 °C |
| C7268 |
Chymostatin |
Inhibits chymotrypsin-like serine proteases such as chymase cathepsins A,B,D and G. Also inhibits some cysteine proteases such as papain. |
10-100 µM |
10 mM in DMSO, stable at -20 °C |
| A9141 |
Antithrombin III |
Inhibits thrombin, kallikreins, plasmin, trypsin and factors Ixa, Xa, and Xia |
equimolar |
Soluble at 10 un/ml in water, prepare stock solutions at neutral pH store at -20 °C |
| D7910 |
3,4-Dichloroisocoumarin |
Broad spectrum serine protease inhibitor |
5-100 µM |
10 mM in DMSO, stable at -20 °C |
| T7254 |
TLCK |
Inhibits trypsin-like serine proteases |
10-100 µM |
Prepare fresh at 10 mM in 1 mM HCl |
| T4376 |
TPCK |
Inhibits chymotrypsin-like serine proteases |
10-100 µM |
10 mM in ethanol, stable at 4 °C |
| D0879 |
DIFP |
Highly toxic cholinesterase inhibitor. Broad spectrum serine protease inhibitor. Hydrolyzes rapidly in aqueous solutions |
0.1 mM |
Prepare in anhydrous isopropanol at 200 mM |
| A6191 |
Antipain |
Inhibits serine proteases such as plasmin, thrombin and trypsin. Also inhibits some cysteine proteases such as calpain and papain. |
1-100 µM |
10 mM in water, stable 1 month at -20 °C |
| M6159 |
a2-Macroglobulin |
Broad spectrum protease inhibitor |
equimolar |
Water soluble, stable at -20 °C |
back to top
Cysteine Protease Inhibitors
| Product Code |
Inhibitor |
Characteristics |
Typical Working Concentration |
Prep. of Stock Solutions |
| E3876 |
N-Ethylmaleimide |
Binds stoichiometrically to SH groups |
equimolar |
Water soluble at >10mg/ml, prepare fresh |
| L2884 |
Leupeptin |
Inhibits trypsin-like serine proteases such as trypsin, chymotrypsin, chymase, pepsin and thrombin. Inhibits selected cysteine proteases such as calpain, cathepsin B, H & L and papain. |
10-100 µM |
10 mM in water, stable 6 months at -20 °C |
| C7268 |
Chymostatin |
Inhibits chymotrypsin-like serine proteases such as chymase cathepsins A,B,D and G. Also inhibits some cysteine proteases such as papain |
10-100 µM |
10 mM in DMSO, stable at -20 °C |
| A6191 |
Antipain |
Inhibits serine proteases such as plasmin, thrombin and trypsin. Also inhibits some cysteine proteases such as calpain and papain. |
1-100 µM |
10 mM in water, stable 1 month at -20 °C |
| M6159 |
a2-Macroglobulin |
Broad spectrum protease inhibitor |
equimolar |
Water soluble, stable at -20 °C |
| P7626 |
PMSF |
Broad spectrum serine protease inhibitor. Also reported to inhibit some cysteine proteases such as papain. |
0.1-1.0 mM |
Prepare fresh in anhydrous ethanol or isopropanol at 200 mM |
back to top
Aspartic Protease Inhibitors
| Product Code |
Inhibitor |
Characteristics |
Typical Working Concentration |
Prep. of Stock Solutions |
| P5318 |
Pepstatin A |
Inhibits aspartic proteases such as renin, chymosin and pepsin |
1 µM |
1mM in methanol or DMSO, stable at -20 °C |
| M6159 |
a2-Macroglobulin |
Broad spectrum protease inhibitor |
equimolar |
Water soluble, stable at -20 °C |
back to top
Metalloproteinase Inhibitors
| Product Code |
Inhibitor |
Characteristics |
Typical Working Concentration |
Prep. of Stock Solutions |
| ED2SS |
EDTA |
Broad spectrum metalloproteinase inhibitor |
1-10 mM |
Very soluble and stable in water |
| P9375 |
1,10-Phenanthroline |
Broad spectrum metalloproteinase inhibitor |
1-10 mM |
200mM in methanol or DMSO, stable at -20 °C |
| R7385 |
Phosphoramidon |
Strong inhibitor of metalloendoproteinases, thermolysin and elastases, but a week inhibitor of collagenase |
1-10 µM |
1 mM in water, stable 1 month at -20 °C |
| B8385 |
Bestatin |
Inhibitor of aminopeptidases |
1-10 µM |
1 mM in methanol, stable 1 month at -20 °C |
| M6159 |
a2-Macroglobulin |
Broad spectrum protease inhibitor |
equimolar |
Water soluble, stable at -20 °C |
back to top
References
- Proteolytic Enzymes: A Practical Approach, R. J. Benyon and J. S. Bond, Eds, pp. 241-249 (1994)
- Handbook of Enzyme Inhibitors, 2nd ed, H. Zollner (1993)
- Sigma Data
back to top back to Protease Inhibitors