|
Caspase
| |
Inhibitors:
|
|
Biotin-Val-Ala-Asp(OMe)-Fluoromethyl ketone, B2057 N-T-Boc-Asp(OMe)-fluoromethyl ketone, B2682 Z-Ala-Glu(OMe)-Val-Asp(OMe)-Fluoromethyl ketone, C8484 Z-Val-Lys(Biotin)-Asp(OMe)-Fluoromethyl ketone, C8609
|
back to top back to Proteolytic Enzymes Guide
Caspase 1
| Product Name |
Product # |
| Caspase 1 human |
C5482 |
| |
Specificity:
|
|
Release of interleukin 1b by specific cleavage at -Asp116-Ala- and -Asp27-Gly- bonds in precursor
|
| |
Synthethic Substrates:
|
|
N-Acetyl-Tyr-Val-Ala-Asp p-nitroanilide, A3831 N-Acetyl-Trp-Glu-His-Asp-p-nitroanilide, A6845 N-Acetyl-Trp-Glu-His-Asp-7-amido-4-methylcoumarin, A0216 N-Acetyl-Tyr-Val-Ala-Asp-7-amido-4-methylcoumarin, A2452 N-Acetyl-Trp-Glu-His-Asp-7-amide-4-trifluoromethylcoumarin, A6720 N-Acetyl-Tyr-Val-Ala-Asp-7-amido-4-trifluoromethylcoumarin, A9965 7-Methoxycoumarin-4-acetyl-Tyr-Val-Ala-Asp-Ala-Pro-(2,4-dinitrophenyl)Lys, M5295
|
| |
Inhibitors:
|
|
N-Acetyl-Trp-Glu-His-Asp-al, A1466 N-Acetyl-Tyr-Val-Ala-Asp chloromethyl ketone, A4211 N-Acetyl-Tyr-Val-Lys(biotinyl)-Asp 2,6-dimethylbenzoyloxymethyl ketone, A4339 Z-Leu-Glu-(OMe)-Val-Asp(OMe)-Fluoromethyl ketone, C9484
|
back to top back to Proteolytic Enzymes Guide
Caspase 2
| Product Name |
Product # |
| Caspase 2 human |
C2854 |
| |
Synthethic Substrates:
|
|
N-Acetyl-Val-Asp-Val-Ala-Asp-p-nitroanilide, A5470 N-Acetyl-Val-Asp-Val-Ala-Asp-7-amido-4-trifluoromethylcoumarin, A5345
|
| |
Inhibitors:
|
|
N-Acetyl-Val-Asp-Val-Ala-Asp-CHO, A2222 N-Cbz-Val-Asp(OMe)-Val-Ala-Asp(OMe)-FMK, C1605
|
back to top back to Proteolytic Enzymes Guide
Caspase 3
| Product Name |
Product # |
| Caspase 3 human |
C1224 |
| |
Specificity:
|
|
Cleaves PARP, ICAD, gelsolin, procaspases 6 and 9
|
| |
Synthethic Substrates:
|
|
N-Acetyl-Asp-Glu-Val-Asp p-nitroanilide, A2559 N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-trifluoromethylcoumarin, A0466 Acetyl-Asp-Glu-Val-Asp-7-amido-4-methylcoumarin, A1086 N-Acetyl-Asp-Gln-Met-Asp-7-amido-4-trifluoromethylcoumarin, A5595 Chromofluor 96, C7605 DFF45/ICAD human, D3815
|
| |
Inhibitors:
|
|
N-AcetylL-Asp-Glu-Val-Asp-al, A0835 N-Acetyl-Ile-Glu-Thr-Asp-al, A1216 N-Acetyl-Glu-Ser-Met-Asp-al, A1341 Biotin-Asp-(OMe)-Glu-(OMe)Val(OMe)-Asp(OMe)-Fluoromethyl ketone, B1807 N-Cbz-Asp(OMe)-Gln-Met-Asp(OMe)-FMK, C0480 Caspase-4 Inhibitor (aldehyde), C0859 Z-VAD-FMK, V116
|
back to top back to Proteolytic Enzymes Guide
Caspase 4
| Product Name |
Product # |
| Caspase 4 human |
C6357 |
| |
Synthethic Substrates:
|
|
N-Acetyl-Tyr-Val-Ala-Asp p-nitroanilide, A3831 N-Acetyl-Leu-Glu-Val-Asp-7-amido-4-trifluoromethylcoumarin, A2099 N-Acetyl-Tyr-Val-Ala-Asp-7-amido-4-methylcoumarin, A2452 N-Acetyl-Tyr-Val-Ala-Asp-7-amido-4-trifluoromethylcoumarin, A9965
|
back to top back to Proteolytic Enzymes Guide
Caspase 5
| Product Name |
Product # |
| Caspase 5 human |
C6482 |
| |
Synthethic Substrates:
|
|
N-Acetyl-Trp-Glu-His-Asp-p-nitroanilide, A6845 N-Acetyl-Trp-Glu-His-Asp-7-amide-4-trifluoromethylcoumarin, A6720
|
| |
Inhibitors:
|
|
N-Acetyl-Trp-Glu-His-Asp-al, A1466
|
back to top back to Proteolytic Enzymes Guide
Caspase 6
| Product Name |
Product # |
| Caspase 6 human |
C4977 |
| |
Synthethic Substrates:
|
|
N-Acetyl-Val-Glu-Ile-Asp-p-nitroanilide, A5220 N-Acetyl-Val-Glu-Ile-Asp-7-amido-4-methylcoumarin, A0341 N-Acetyl-Val-Glu-His-Asp-7-amido-4-trifluoromethylcoumarin, A2097 N-Acetyl-Val-Glu-Ile-Asp-7-amido-4-trifluoromethylcoumarin, A5095 Caspase 6 Assay Kit, Fluorimetric, CASP-6-F
|
| |
Inhibitors:
|
|
N-Acetyl-Val-Glu-Ile-Asp-al, A6339 Biotin-Asp-(OMe)-Glu-(OMe)Val(OMe)-Asp(OMe)-Fluoromethyl ketone, B1807 N-Cbz-Asp(OMe)-Gln-Met-Asp(OMe)-FMK, C0480 N-CBZ-Val-Glu-(OMe)-Ile-Asp(OMe)-FMK, C1730
|
back to top back to Proteolytic Enzymes Guide
Caspase 7
| Product Name |
Product # |
| Caspase 7 human |
C2979 |
| |
Synthethic Substrates:
|
|
N-Acetyl-Asp-Glu-Val-Asp p-nitroanilide, A2559 N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-trifluoromethylcoumarin, A0466 Acetyl-Asp-Glu-Val-Asp-7-amido-4-methylcoumarin, A1086
|
| |
Inhibitors:
|
|
Biotin-Asp-(OMe)-Glu-(OMe)Val(OMe)-Asp(OMe)-Fluoromethyl ketone, B1807 N-Cbz-Asp(OMe)-GLu(OMe)-Val-Asp(OMe)-FMK, C0605
|
back to top back to Proteolytic Enzymes Guide
Caspase 8
| |
Specificity:
|
|
Cleaves procaspases 3,6, and 7
|
| |
Synthethic Substrates:
|
|
N-Acetyl-Ile-Glu-Thr-Asp-p-nitroanilide, A9968 Caspase 8 Colorimetric Kit, CASP-8-C N-Acetyl-Ala-Glu-Val-Asp-7-amido-4-trifluoromethylcoumarin, A0973 N-Acetyl-Ile-Glu-Thr-Asp-7-Amido-4-methylcoumarin, A4188 N-Acetyl-Leu-Glu-Thr-Asp-7-Amido-4-trifluoromethylcoumarin, A6095 Z-Ile-Glu-Thr-Asp 7-amido-4-trifluoromethylcoumarin, C5599 Chromofluor 96, C7605 Caspase 8 Fluorimetric Kit, CASP-8-F
|
| |
Inhibitors:
|
|
N-Acetyl-Ile-Glu-Thr-Asp-al, A1216 Biotin-Asp-(OMe)-Glu-(OMe)Val(OMe)-Asp(OMe)-Fluoromethyl ketone, B1807 N-Cbz-Ile-Glu(OMe)-Thr-Asp(OMe), C1230 Z-Leu-Glu-(OMe)-Thr-Asp-(OMe)-Fluoromethyl ketone, C8734
|
back to top back to Proteolytic Enzymes Guide
Caspase 9
| Product Name |
Product # |
| Caspase 9 human |
C8726 |
| |
Specificity:
|
|
Cleaves procaspases 3,6, and 7
|
| |
Synthethic Substrates:
|
|
N-Acetyl-Leu-Glu-His-Asp-p-nitroaniline, A5970 N-Acetyl-Val-Glu-His-Asp-7-amido-4-trifluoromethylcoumarin trifluoroacetate salt, A2097 N-Acetyl-Leu-Glu-His-Asp-7-amido-4-trifluoromethylcoumarin, A5845 Chromofluor 96, C7605
|
| |
Inhibitors:
|
|
N-Cbz-Leu-Glu(OMe)-His-Asp(OMe)-FMK, C1355
|
back to top back to Proteolytic Enzymes Guide
Caspase 10
| Product Name |
Product # |
| Caspase 10 human, recombinant, expressed in Escherichia coli >90% (SDS-PAGE) Lyophilized powder 8,000 units/mg protein |
C7982 |
| Caspase 10 human, recombinant, expressed in Escherichia coli >90% (SDS-PAGE) Lyophilized powder 8,000 units/mg protein (Bradford) |
C6607 |
| |
Specificity:
|
|
Cleaves procaspases 3, 4, 6, 7, 8 and 9
|
| |
Inhibitors:
|
|
Biotin-Asp-(OMe)-Glu-(OMe)Val(OMe)-Asp(OMe)-Fluoromethyl ketone, B1807
|
back to top back to Proteolytic Enzymes Guide
Caspase 13
| |
Inhibitors:
|
|
Z-Leu-Glu(OMe)-Glu(OMe)-Asp(OMe)-Fluoromethyl ketone, C8859
|
back to top back to Proteolytic Enzymes Guide
Cathepsin B, E.C. 3.4.22.1
| Product Name |
Product # |
| Cathepsin B from bovine spleen |
C6286 |
| Cathepsin B from human liver |
C8571 |
| Cathepsin B from human placenta |
C0150 |
| |
Specificity:
|
|
Hydrolysis of proteins with broad specificity for peptide bonds. Preferentially cleaves -Arg-Arg bonds in small molecule substrates (thus differing from cathepsin L). In addition to being an endopeptidase, shows peptidyl-dipeptidase activity, liberating C-terminal dipeptides
|
| |
Synthethic Substrates:
|
|
Na-Benzoyl-L-arginine 4-methoxy-beta-naphthylamide hydrochloride, B0885 Z-Val-Lys-Lys-Arg-4-methoxy-beta-naphthylamide trihydrochloride, C2772 Z-Phe-Arg 4-methoxy-beta-naphthylamide hydrochloride, C3282 Z-Arg-Arg-7-amido-4-Z-RR-AMC-methylcoumarin hydrochloride, C5429 Z-Arg-Arg 4-methoxy-beta-naphthylamide acetate salt, C5520 Z-Ala-Arg-Arg 4-methoxy-beta-naphthylamide acetate salt, C8536 N-Succinyl-Leu-Leu-Val-Tyr-7-Amido-4-Methylcoumarin, S6510 Hippuryl-Arg, H2508
|
| |
Inhibitors:
|
|
N-Acetyl-Leu-Leu-Met-al, A6060 N-Acetyl-Leu-Leu-Norleu-al, A6185 Antipain, A6191 Biotin-Phe-Ala-Fluoromethyl ketone, B1932 Cystatin, C0408 CA-074, C5732 CA-074 Methyl ester, C5857 Chymostatin, C7268 Cystatin, C8917 Z-Phe-Phe-Fluoromethyl ketone, C9109 E-64, E3132 Leupeptin, L2023 MeOSuc-Phe-homo-Phe fluoromethyl ketone, M5314 a2-Macroglobulin, M6159 Procathepsin B, rat, fragment 26-50, P0350 Procathepsin B, rat, fragment 36-50, P0475
|
back to top back to Proteolytic Enzymes Guide
Cathepsin C, E.C. 3.4.14.1
| Product Name |
Product # |
| Cathepsin C from bovine spleen |
C8511 |
| |
Specificity:
|
|
Release of an N-terminal dipeptide, Xaa-Xbb Xcc-, except when Xaa is Arg or Lys, or Xbb or Xcc is Pro
|
| |
Synthethic Substrates:
|
|
Gly-Phe p-nitroanilide, G0142 Gly-Phe beta-naphthylamide, G4255 Gly-Phe beta-naphthylamide, G9512 Ser-Tyr beta-naphthylamide, S5628
|
| |
Inhibitors:
|
|
Iodoacetic Acid, I2512
|
back to top back to Proteolytic Enzymes Guide
Cathepsin D, E.C. 3.4.23.5
| Product Name |
Product # |
| Cathepsin D from bovine spleen |
C3138 |
| Cathepsin D from human liver |
C8696 |
| |
Specificity:
|
|
Specificity similar to, but narrower than, that of pepsin A. Does not cleave the Gln4-His bond in B chain of insulin
|
| |
Synthethic Substrates:
|
|
Boc-Phe-Ala-Ala-p-Nitro-Phe-Phe-Val-Leu 4-hydroxymethylpyridine ester, B1644 N-Acetyl-Arg-Gly-Phe-Phe-Pro-7-amido-4-methylcoumarin, C6358 N-Acetyl-Arg-Gly-Phe-Phe-Pro-7-amido-4-trifluoromethylcoumarin, C6983 N-Acetyl-Arg-Gly-Phe-Phe-Pro 4-methoxy-2-naphthylamide, C7108 7-Methoxycoumarin-4-Acetyl-Gly-Lys-Pro-Ile-Leu-Phe-Phe-Arg-Leu-Lys(DNP)-D-Arg-amide, M0938
|
| |
Inhibitors:
|
|
Antipain, A6191 Chymostatin, C7268 Pepstatin A, P5318
|
back to top back to Proteolytic Enzymes Guide
Cathepsin G, E.C. 3.4.21.20
| Product Name |
Product # |
| Cathepsin G from human leukocytes |
C4428 |
| Cathepsin G from human sputum |
C5108 |
| |
Specificity:
|
|
Specificity similar to chymotrypsin C
|
| |
Synthethic Substrates:
|
|
N-Methoxysuccinyl-Ala-Ala-Pro-Met p-nitroanilide, M7771 N-Succinyl-Ala-Ala-Pro-Phe p-nitroanilide, S7388 N-Succinyl-Gly-Gly-Phe-7-amido-4-trifluoromethylcoumarin, S4689 N-Succinyl-Gly-Gly-Phe-4-methoxy-2-naphthylamide, S4814 N-Succinyl-Ala-Ala-Pro-Phe thiobenzyl ester, S6518 N-Succinyl-Phe-Leu-Phe thiobenzyl ester, S6643
|
| |
Inhibitors:
|
|
a1-Antchymotrypsin, A9285 Chymostatin, C7268 Z-Gly-Leu-Phe-Chloromethyl ketone, C9984 Diisopropyl fluorophosphate, D0879 3,4-Dichloroisocoumarin, D7910 N-Acetyl-eglin C, E7888
|
back to top back to Proteolytic Enzymes Guide
Cathepsin H, E.C. 3.4.22.16
| Product Name |
Product # |
| Cathepsin H from human liver |
C8821 |
| |
Specificity:
|
|
Hydrolysis of proteins, acting as an aminopeptidase (notably, cleaving Arg bonds) as well as an endopeptidase
|
| |
Synthethic Substrates:
|
|
L-Arginine-7-amido-4-methylcoumarin hydrochloride, A2027
|
back to top back to Proteolytic Enzymes Guide
Cathepsin L, E.C. 3.4.22.15
| Product Name |
Product # |
| Cathepsin L from human liver |
C6854 |
| |
Specificity:
|
|
Similar to that of papain. As compared to cathepsin B, cathepsin L exhibits higher activity towards protein substrates and no peptidyl-dipeptidase activity
|
| |
Synthethic Substrates:
|
|
Z-Phe-Arg 4-methoxy-beta-naphthylamide hydrochloride, C3282
|
| |
Inhibitors:
|
|
N-Acetyl-Leu-Leu-Norleu-al, A6185
|
back to top back to Proteolytic Enzymes Guide
Chymopapain, E.C. 3.4.22.6
| Product Name |
Product # |
| Chymopapain from papaya latex |
C8526 |
| |
Specificity:
|
|
Similar to that of papain
|
| |
Synthethic Substrates:
|
|
BAEE, B4500
|
back to top back to Proteolytic Enzymes Guide
|