Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Metabolomics > Enzyme Explorer > Learning Center > Proteolytic Enzymes > Proteolytic Enzymes IV
Enzyme Explorer
Proteolytic Enzymes IV
Ficin, E.C. 3.4.22.3
Product Name
Product # 
Ficin from fig tree latex, Saline suspension minimum 1.5 units/mg protein (biuret) F4125
Ficin from fig tree latex, Lyophilized powder minimum 1 units/mg protein F6008
Ficin from fig tree latex, powder minimum 0.1 unit/mg solid F4165
Specificity:
Similar to that of papain
Inhibitors:
Cystatin, C8917
E-64, E3132
Iodoacetic Acid, I2512
N-a-p-Tosyl-L-lysine chloromethyl ketone Hydrochloride, T7254
N-a-p-Tosyl-L-phenylalanine chloromethyl ketone Hydrochloride, T4376
N-Ethylmaleimide, E3876
back to top
back to Proteolytic Enzymes Guide
Furin, E.C. 3.4.21.75
Product Name
Product # 
Furin F2677
Specificity:
Release of mature proteins from their proproteins by cleavage of -Arg-Xaa-Yaa-Arg bonds, where Xaa can by any amino acid and Yaa is Arg or Lys. Releases albumin, complement component C3 and von Willebrand factor from their respective precursors
back to top
back to Proteolytic Enzymes Guide
Granzyme A, E.C. 3.4.21.78
Specificity:
Hydrolysis of proteins, including fibronectin, type IV collagen and nucleolin. Preferential cleavage: -Arg-Xaa, -Lys-Xaa >> -Phe-Xaa in small molecule substrates
Synthethic Substrates:
N-CBZ-GLY-PRO-ARG 7-AMIDO-4-METHYLCOUMARIN, C0855
N-CBZ-GLY-PRO-ARG 7-AMIDO-4-TRIFLUOROMETHYLCOUMARIN, C0980
back to top
back to Proteolytic Enzymes Guide
Granzyme B
Specificity:
Preferential cleavage: -Asp-Xaa >> -Asn-Xaa > -Met-Xaa, -Ser-Xaa
Synthethic Substrates:
Z-Gly-Pro-Arg 4-methoxy-2-naphthylamide, C1105
N-Acetyl-Ile-Glu-Thr-Asp-7-Amido-4-methylcoumarin, A4188
N-Acetyl-Ile-Glu-Pro-Asp-7-amido-4-methylcoumarin, A6220
N-Acetyl-Ile-Glu-Pro-Asp-p-nitroaniline, A6470
N-Acetyl-Ile-Glu-Thr-Asp-p-nitroanilide, A9968
Z-Ile-Glu-Thr-Asp 7-amido-4-trifluoromethylcoumarin, C5599
Inhibitors:
Antipain, A6191
3,4-Dichloroisocoumarin, D7910
back to top
back to Proteolytic Enzymes Guide
HIV Protease, E.C. 3.4.23.16
Product Name
Product # 
Protease from HIV-1 P7338
Specificity:
Specific for a P1 residue that is hydrophobic, and P1' variable, but often Pro
Synthethic Substrates:
HIV Protease Substrate I , H6660
HIV Protease Substrate V, H5410
HIV Protease Substrate VIII, H5785
Anthranilyl-HIV Protease Substrate V, H6035
Anthranilyl-HIV Protease Substrate VI, H6160
back to top
back to Proteolytic Enzymes Guide
IGase
Product Name
Product # 
Endoproteinase Pro-Pro-Y-Pro E2528
Specificity:
Hydrolyzes Pro-Pro-|-Y-Pro
back to top
back to Proteolytic Enzymes Guide
Kallikrein tissue, E.C. 3.4.21.35
Product Name
Product # 
Endoproteinase Arg-C murine submaxillary gland P6056
Specificity:
Preferential cleavage of Arg-Xaa bonds in small molecule substrates. Highly selective action to release kallidin (lysyl-bradykinin) from kininogen involves hydrolysis of Met-Xaa or Leu-Xaa . The rat enzyme is unusual in liberating bradykinin directly from autologous kininogens by cleavage at two Arg bonds
Synthethic Substrates:
N-Benzoyl-Pro-Phe-Arg-p-nitroanilide hydrochloride, B2133
Z-Phe-Arg 7-amido-4-methylcoumarin hydrochl, C9521
Kininogen, low molecular weight from human plasma, K3628
D-Val-Leu-Arg 7-amido-4-methylcoumarin, V2763
D-Val-Leu-Arg 7-amido-4-trifluoromethylcoumarin, V2888
Inhibitors:
Aprotinin, A1153
Antithrombin III, Human, A2221
Antithrombin III, Bovine, A9141
Benzamidine Hydrochloride, B6506
Diisopropyl fluorophosphate, D0879
3,4-Dichloroisocoumarin, D7910
Leupeptin, L2023
back to top
back to Proteolytic Enzymes Guide
Leucine Aminopeptidase (General)
Inhibitors:
Actinonin, A6671
Bestatin Hydrochloride, B8385
back to top
back to Proteolytic Enzymes Guide
Leucine aminopeptidase, cytosol, E.C. 3.4.11.1
Specificity:
Release of an N-terminal amino acid, Xaa Xbb-, in which Xaa is preferably Leu, but may be other amino acids including Pro although not Arg or Lys, and Xbb may be Pro. Amino acid amides and methyl esters are also readily hydrolysed, but rates on arylamides are exceedingly low
Synthethic Substrates:
L-Leucine p-Nitroanilide, L9125
N-Succinyl-Ala-Ala-Pro-Phe-7-amido-4-methylcoumarin, S9761
Inhibitors:
Amastatin Hydrochloride, A1276
(2S,3R)-3-Amino-2-hydroxy-4-(4-nitro«-»phenyl)butanoyl-L-leucine Hydrochloride, A5921
Actinonin, A6671
Bestatin Hydrochloride, B8385
Diethylenetriaminepentaacetic acid Free Acid, D6518
3,4-Dichloroisocoumarin, D7910
EDTA, ED2SS
EGTA, E4378
1,10-Phenanthroline Monohydrate, P9375
back to top
back to Proteolytic Enzymes Guide
Leucine aminopeptidase, microsomal, E.C. 3.4.11.2
Product Name
Product # 
Leucine Aminopeptidase, microsomal from porcine kidney, Type IV-S Ammonium sulfate suspension 10-40 units/mg protein (Bradford) L5006
Leucine Aminopeptidase, microsomal from porcine kidney, Type VI-S Lyophilized powder 15-25 units/mg protein (biuret) L6007
Leucine Aminopeptidase, microsomal from porcine kidney, suitable for protein sequencing Lyophilized powder L9776
Specificity:
Release of an N-terminal amino acid, Xaa Xbb- from a peptide, amide or arylamide. Xaa is preferably Ala, but may be most amino acids including Pro (slow action). When a terminal hydrophobic residue is followed by a prolyl residue, the two may be released as an intact Xaa-Pro dipeptide
Synthethic Substrates:
L-Leucine p-Nitroanilide, L9125
N-Succinyl-Ala-Ala-Pro-Phe-7-amido-4-methylcoumarin, S9761
Inhibitors:
Amastatin Hydrochloride, A1276
Actinonin, A6671
Bestatin Hydrochloride, B8385
back to top
back to Proteolytic Enzymes Guide
Matrix metalloprotease
Product Name
Product # 
Matrix Metalloproteinase-7 from human kidney cells M4565
MMP Control-4 M4815
MMP Control-5 M5065
MMP Control-6 M4940
Specificity:
Specificity is MMP type specific
Synthethic Substrates:
7-Methoxycoumarin-4-acetyl-Pro-Leu-Gly-Leu-b-(2,4-dinitrophenylamino)Ala-Ala-Arg amide, M6412
Inhibitors:
Actinonin, A6671
4-Aminobenzoyl-Gly-Pro-D-Leu-D-Ala hydroxamic acid, A4336
Dichloromethylenediphosphonic acid disodium salt, D4434
Isoamylphosphonyl-Gly-Pro-Ala dipotassium salt, I7029
N-[(2S)-(Methoxycarbonylmethyl)-4-methylpentanoyl]-L-tryptophan-methylamide, M5939
Z-Pro-Leu-Gly hydroxamate, C8537
Pro-Leu-Gly hydroxamate hydrochloride, P5298
Tissue Inhibitor of Metalloproteinase-1 from human fibroblasts, T8947
Tissue Inhibitor of Metalloproteinase-4 from human fibroblasts, T9322
back to top
back to Proteolytic Enzymes Guide
Methionine Aminopeptidase, E.C. 3.4.11.18
Product Name
Product # 
Methionine Aminopeptidase Pyrococcus furiosus M6435
Specificity:
Hydrolysis of N-terminal methionine
back to top
back to Proteolytic Enzymes Guide
Neutrase
Product Name
Product # 
Protease from Bacillus amyloliquefaciens P1236
back to top
back to Proteolytic Enzymes Guide
Papain, E.C. 3.4.22.2
Product Name
Product # 
Papain from papaya latex, Buffered aqueous suspension 16-40 units/mg protein P3125
Papain Agarose preparation from papaya latex P4406
Papain from papaya latex, Crude powder 1.5-3.5 units/mg solid P3375
Papain from papaya latex, 0.5-2 units/mg solid P3250
Papain from papaya latex, minimum 10 units/mg protein (E1%/280) P4762
Papain from papaya latex P5306
Specificity:
Hydrolysis of proteins with broad specificity for peptide bonds, but preference for an amino acid bearing a large hydrophobic side chain at the P2 position. Does not accept Val in P1'
Synthethic Substrates:
Na-Benzoyl-L-arginine 7-amido-4-methylcoumarin Hydrochloride, B7260
Inhibitors:
Antipain, A6191
Chymostatin, C7268
Cystatin, C8917
3,4-Dichloroisocoumarin, D7910
E-64, E3132
Ebselen, E3520
Gly-Gly-Tyr-Arg, G5386
Leupeptin, L2023
a2-Macroglobulin, M6159
N-a-p-Tosyl-L-lysine chloromethyl ketone Hydrochloride, T7254
N-a-p-Tosyl-L-phenylalanine chloromethyl ketone Hydrochloride, T4376
back to top
back to Proteolytic Enzymes Guide
Pepsin, E.C. 3.4.23.1
Product Name
Product # 
Pepsin porcine gastric mucosa, Lyophilized powder 3,200-4,500 units/mg protein P6887
Pepsin porcine gastric mucosa, Lyophilized powder 2,500-3,500 units/mg protein (E1%/280) P7012
Pepsin porcine gastric mucosa, powder 800-2,500 units/mg protein P7000
Pepsin porcine gastric mucosa, powder 600-1,800 units/mg protein P7125
Pepsin Agarose from porcine gastric mucosa P0609
Specificity:
Preferential cleavage: hydrophobic, preferably aromatic, residues in P1 and P1' positions. Cleaves Phe1 Val, Gln4 His, Glu13 Ala, Ala14 Ala, Ala14 Leu, Leu15 Tyr, Tyr16 Leu, Gly23 Phe, Phe24 Phe and Phe25 Tyr bonds in the B chain of insulin
Inhibitors:
Pepstatin A, P5318
back to top
back to Proteolytic Enzymes Guide