Enzyme Explorer
Substrates Procedures
Substrate Product # 
Substrate Procedure
C0403
Casein fluorescein isothiocyanate from bovine milk, Type I
C0528
Casein fluorescein isothiocyanate from bovine milk, Type III Fluorometric Protease Assay (PDF 75.3 KB)
C3777
Casein fluorescein isothiocyanate from bovine milk, Type II Fluorometric Protease Assay (PDF 75.3 KB)
C7078
Casein bovine milk, Technical Grade Colorimetric Protease Assay (PDF 17.7 KB), Instructional Video of Assay
C9801
Casein, Dimethylated Colorimetric Protease Assay: Cabacungan, J.C., et al., Anal. Biochem., 124, 272 (1982)
E0502
Elastin-Congo Red Colorimetric Protease Assay at 495 nm: Rust, L., et. al., Meth. Enzymol., 235, 557-559 (1994) and Shotton, D. M., Meth. Enzymol., XIX, 113-117 (1970)
E1500
Elastin-Orcein Colorimetric Protease Assay at 570 nm: Sachar, L. A., et. al., Proc. Soc. Exptl. Bio. Med., 90, 323 (1955)
G7765
Gelatin Use in Zymograms: Kleiner, D.E, and Stetler-Stevenson, W.G., Anal. Biochem., 218, 325-329 (1994) and Heussen, C. and Dowdle, E.B., Anal. Biochem., 102, 196-202 (1980)
H2625
Hemoglobin, Bovine Colorimetric Protease Assay
H6268
Remazol Brilliant Blue R Colorimetric Protease Assay at 595 nm: Rinderknecht, H., et al., Clin. Chem. Acta, 21, 197 (1968)
H9891
Hemoglobin, N,N-Dimethylated Colorimetric Protease Assay: Cabacungan, J.C., et al., Anal. Biochem., 124, 272 (1982)
back to Proteolytic Enzymes Guide