Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Structural Proteins
Aggrecan
Aggrecan is the major structural proteoglycan found in the extracellular matrix of cartilage. It has a molecular mass >2,500 kDa. The core protein (210-250 kDa) has 100-150 glycosaminoglycan (GAG) chains attached to it. The majority of the GAG chains are chondroitin/dermatan sulfate with the remainder being keratan sulfate. This structural molecule produces a rigid, reversibly deformable gel that resists compression. It combines with hyaluronic acid to form very large macromolecular complexes. Addition of small amounts (0.1-2% w/w) of hyaluronic acid to a solution of aggrecan (2 mg/ml) results in the formation of a complex with an increased hydrodynamic volume and in a significant increase (30-40%) in the relative viscosity of the solution. Aggrecan is a critical component for cartilage structure and the function of joints. The synthesis and degradation of aggrecan are being investigated for their roles in cartilage deterioration during joint injury, disease, and aging. It contains three globular domains, G1, G2, and G3 that are involved in aggregation, hyaluronan binding, cell adhesion, and chondrocyte apoptosis.
Aggrecan Illustration
Aggrecan Products
back to Structural Proteins