|
Structural Proteins
Myelin Basic Protein
|
|
|
Myelin basic protein (MBP) comprises about 20% of the myelin sheath in neuronal axons. MBP is commonly used to induce autoimmune encephalomyelitis in animal models. MBP is unique in its high degree of post-translational modifications including multiple phosphorylation sites, deamidation of glutamine and acetylation of alanine as well as deimination of arginine to yield citrulline and methylation of arginine.
Myelin Basic Protein Products
back to Structural Proteins
|