|
Structural Proteins
Tropomyosin
|
|
|
Tropomyosin together with troponin, regulate the binding of myosin to actin. Tropomyosin is a dimeric coiled-coil protein forming continuous polymers along the myosin-binding groove of actin. Varoius tropomyosin isoforms help to confer structure and function to actins in the cytoskeleton and in striated muscle function.
Tropomyosin Products
back to Structural Proteins
|