Sigma-Aldrich
Life Science > Metabolomics > Enzyme Explorer > Learning Center > Structural Proteins > Tropomyosin
Structural Proteins
Tropomyosin
Tropomyosin together with troponin, regulate the binding of myosin to actin. Tropomyosin is a dimeric coiled-coil protein forming continuous polymers along the myosin-binding groove of actin. Varoius tropomyosin isoforms help to confer structure and function to actins in the cytoskeleton and in striated muscle function. 
Tropomyosin Illustration
Tropomyosin Products
back to Structural Proteins