Sigma-Aldrich
Life Science > Metabolomics > Enzyme Explorer > Learning Center > Structural Proteins > Troponin
Structural Proteins
Troponin
Troponin together with tropomyosin, regulate the binding of myosin to actin. Troponin is a trimeric protein composed of Troponin subunits I, C and T. Troponin C binds calcium ions, Troponin T binds to tropomyosin and troponin I binds to actin. This calcium-dependent interaction serves to position the tropomyosin along the actin filament during muscle contraction. Troponins are used as diagnostic biomarkers for cardiac injury. 
Troponin Illustration
Troponin Products
back to Structural Proteins