Sigma has the largest selection of lectins and lectin conjugates available for the life scientist. Over 100 products from over 35 sources make Sigma the single best source for your lectin requirements.
Features and Benefits:
Backed by quality and service you’ve come to expect, Sigma offers lectins for alLyour life science applications, including;
- HistochemicaLStudies
- Lymphocyte Subpopulations Studies
- CelLFractionation
|
- MitogeniCStimulation
- Blood Group Typing
|
Link To: TaxonomiCSource Listing Conjugate Details Product List (includes conjugates, specificity, mitogeniCactivity, and more)
TaxonomiCSource Listing:
| Common Name |
TaxonomiCName |
|
Common Name |
TaxonomiCName |
| *Abrin |
Abrus precatorius |
Jimson weed |
Datura stramonium |
| Asparagus pea |
Tetragonolobus purpureas |
*Labumum, Scotch |
Labumum alpinum |
| *Avocado |
Perseau americana |
Lentil |
Lens culinaris |
| *Bitter pear melon |
Momordica charantia |
Lotus |
Tetragonolobus purpureas |
| Broad bean |
Vicia faba |
Mistletoe, European |
Viscum album |
| Camels foot tree |
Bauhinia purpurea |
*Mung bean |
Vigna radiata |
| Castor bean |
Ricinus communis |
Mushroom |
Agaricus bisporus |
| Chick pea |
Cicer arietinum |
Osage orange |
Maclura pomifera |
| *Cobra, Mozambique |
Naja mocambique mocambique |
*Pagoda tree |
Sophora japonica |
| *Cobra, Thailand |
Naja naja kaouthia |
Pea, garden |
Pisum sativum |
| Con A |
Canavalia ensiformis |
Peanut |
Arachis hykpogaea |
| Concanavalin A |
Canavalia ensiformis |
Pokeweek |
Phytolacca americana |
| CoraLtree |
Erythrina cristagalli |
Potato |
Solanum tuberosum |
| CoraLtree, Israel |
Erythrina corallodendron |
Red kidney bean |
Phaseolus vulgaris |
| Daffodil |
Narcissus pseudonarcissus |
*Red marine algae |
Ptilota plumosa |
| Eel |
Anguilla anguilla |
Roman snail |
Helix pomatia |
| Elder |
Sambucus nigra |
Scarlet runner bean |
Phaseolus coccineus |
| Fava bean |
Vicia faba |
Siberian pea tree |
Caragana arborescens |
| Furze |
Ulex europaeus |
Snail, garden |
Helix aspersa |
| Gorse |
Ulex europaeus |
Snowdrop |
Galanthus nivalis |
| Green marine algae |
Codium fragile |
Soybean |
Glycine max |
| Hairy vetch |
Vicia villosa |
Spindle tree |
Euonymus eruopaeus |
| Horse gram |
Dolichos biflorus |
*Sweet pea |
Lathyrus odoratus |
| Horseshoe crab |
Limulus polyphemus |
Thorn apple |
Datura stramonium |
| Jacalin |
Artocarpus integrifolia |
Tomato |
Lycopersicon esculentum |
| Jack bean |
Canavalia ensiformis |
Wheat germ |
Triticum vulgaris |
| Japanese wisteris |
Wisteria floribunda |
Winged bean |
Psophocarpus tetragonolobus |
| *Jequity bean |
Abrus precatorius |
Winged pea |
Tetragonolobus purpureas |
| *Not currently available, please click here to inquire. |
Top of Page |
Conjugate Details:
Sigma has the largest selection of lectins and lectin conjugates available for the life scientist. Over 100 products from over 35 naturaLsources. Conjugates include:
• Agarose • FITC• Peroxidase • Biotin • TRITC• Gold
Many lectins are available as conjugates. Conjugation does not alter the specificity of the lectin. The degree of conjugation is expressed as 1) a mole dye per mole lectin basis 2) a unit enzyme activity per mg protein or 3) a mg protein per mLpacked geLbasis. For colloidaLgold conjugates, the activity of the lectin is determined by a dot blot assay using an appropriate glycoprotein. Agglutination activity is expressed in microgram per mLand is determined from seriaLdilution studies of a 1 mg per mLsolution. This activity is the lowest concentration of lectin to agglutinate a 2% suspension of appropriate erythrocytes after 1 hour incubation at 25 0C. Top of Page
Lectins Product List:
|
Prod. No.: |
LECTIN: |
Mol. Wt. (kDa): |
Subunits: |
Specificity Blood Group: |
Specificity Sugar: |
Mitogenic Activity: |
Conjugates: |
| L5640 |
Agaricus bisporus |
58.5 |
— |
— |
ß-gal(1->3)galNAc |
|
|
| L4141 |
Anguilla anguilla |
40 |
2 |
H |
a-L-Fuc |
|
|
| L0881 |
Arachis hypogaea |
120 |
4 |
T |
ß-gal(1->3)galNAc |
|
|
| L7759 |
conjugate |
|
|
Peroxidase |
| L6135 |
conjugate |
|
|
Biotin |
| L7381 |
conjugate |
|
|
FITC |
| L3766 |
conjugate |
|
|
TRITC |
| L6646 |
conjugate |
|
|
Agarose |
| L3515 |
Artocarpus integrifolia |
42 |
4 |
T |
a-gal->OMe |
(+) |
|
| L4650 |
conjugate |
|
|
Peroxidase |
| L5147 |
conjugate |
|
|
Agarose |
| Bandeiraea Simplicifolia |
Top of Page |
| L2380 |
BS-I |
114 |
4 |
A, B |
a-gal, a-galNAc |
|
|
| L3759 |
conjugate |
|
|
Biotin |
| L9381 |
conjugate |
|
|
FITC |
| L5264 |
conjugate |
|
|
TRITC |
| L1509 |
BS-I-A4 |
114 |
4 |
A |
a-galNAc |
|
|
| L0890 |
conjugate |
|
|
FITC |
| L3019 |
BS-I-B4 |
114 |
4 |
B |
a-gal |
|
|
| L5391 |
conjugate |
|
|
Peroxidase |
| L2140 |
conjugate |
|
|
Biotin |
| L2895 |
conjugate |
|
|
FITC |
| L6013 |
Bauhinia purpurea |
195 |
4 |
— |
ß-gal(1->3)galNAc |
(+) |
|
| L9637 |
Caragana arborescens |
60;120 (c) |
2;4 |
— |
galNAc |
|
|
| L3141 |
Cicer arietinum |
44 |
2 |
— |
fetuin |
|
|
| L2638 |
Codium fragile |
60 |
4 |
— |
galNAc |
|
|
| C7275 |
Concanavalin A |
102 |
4 |
— |
a-man, a-glc |
(+) |
|
| L6397 |
conjugate |
|
|
Peroxidase |
| C2272 |
conjugate |
|
|
Biotin |
| C7642 |
conjugate |
|
|
FITC |
| C6904 |
conjugate |
|
|
Agarose |
| C9017 |
conjugate |
|
|
Sepharose |
| L5021 |
conjugate |
|
|
Gold, 10nm |
| L3642 |
conjugate |
|
|
Gold, 20nm |
| C7898 |
conjugate |
|
|
Ferritin |
| L3885 |
Succinyl-Concanavalin A |
51 |
2 |
— |
a-man, a-glc |
(+) (d) |
|
| L9385 |
conjugate |
|
|
FITC |
| L2766 |
Datura stramonium |
86 |
2(a&ß)a |
— |
(glcNAc)2 |
|
|
| L2785 |
Dolichos biflorus |
140 |
4 |
A1 |
a-galNAc |
|
|
| L1287 |
conjugate |
|
|
Peroxidase |
| L6533 |
conjugate |
|
|
Biotin |
| L9142 |
conjugate |
|
|
FITC |
| L9658 |
conjugate |
|
|
TRITC |
| L9894 |
conjugate |
|
|
Agarose |
| L2142 |
Erythrina corallodendron |
60 |
2 |
— |
ß-gal(1->4)glcNAc |
(+) |
|
| L5390 |
Erythrina cristagalli |
56.8 |
2(a&ß)a |
— |
ß-gal(1->4)glcNAc |
|
|
| L7400 |
Euonymus europaeus |
166 |
4(a&ß)a |
B, H |
a-gal(1->3)gal |
(+) |
|
| L8725 |
Galanthus nivalis |
52 |
4 |
(h) |
non-reduc. D-man |
|
|
| L8775 |
conjugate |
|
|
Agarose |
| L1395 |
Glycine max |
110 |
4 |
— |
galNAc |
(+) (b) |
|
| L2650 |
conjugate |
|
|
Peroxidase |
| L4511 |
conjugate |
|
|
TRITC |
| L1105 |
conjugate |
|
|
Agarose |
| L6635 |
Helix aspersa |
79 |
— |
A |
galNAc |
|
|
| L8764 |
conjugate |
|
|
Biotin |
| L3382 |
Helix pomatia |
79 |
6 |
A |
galNAc |
|
|
| L6387 |
conjugate |
|
|
Peroxidase |
| L6512 |
conjugate |
|
|
Biotin |
| L1034 |
conjugate |
|
|
FITC |
| L1261 |
conjugate |
|
|
TRITC |
| L9267 |
Lens culinaris |
49 |
2 |
— |
a-man |
(+) |
|
| L4143 |
conjugate |
|
|
Biotin |
| L9262 |
conjugate |
|
|
FITC |
| L0511 |
conjugate |
|
|
Sepharose |
| L2263 |
Limulus polyphemus |
400 |
18 |
— |
NeuNAc |
|
|
| L2886 |
Lycopersicon esculentum |
71 |
— |
— |
(glcNAc)3 |
(+) (e) |
|
| L0651 |
conjugate |
|
|
Biotin |
| L0401 |
conjugate |
|
|
FITC |
| L8025 |
Maackia amurensis |
130 |
2(a&ß) |
O |
sialiCacid |
(+) |
|
| L6141 |
Maclura pomifera |
40-43 |
2(a&ß)a |
— |
a-gal,a-galNAc |
|
|
| L4401 |
conjugate |
|
|
Peroxidase |
| L2013 |
conjugate |
|
|
Biotin |
| L5650 |
Narcissus pseudonarcissus |
26 |
2 |
(h) |
a-D-man |
|
|
| L3138 |
Phaseolus coccineus |
112 |
4 |
— |
— |
|
|
| L4389 |
conjugate |
|
|
TRITC |
| Phaseolus Vulgaris |
Top of Page |
| L8629 |
PHA-E |
128 |
4 |
— |
oligosaccharide |
(+) |
|
| L6139 |
conjugate |
|
|
TRITC |
| L2769 |
PHA-L |
126 |
4 |
— |
oligosaccharide |
(+) |
|
| L8754 |
PHA-P |
|
| L2646 |
PHA-M |
|
| L9379 |
Phytolacca americana |
32f |
— |
— |
(glcNAc)3 |
(+) |
|
| L2882 |
conjugate |
|
|
Agarose |
| L5380 |
Pisum sativum |
49 |
4(a&ß)a |
— |
a-man |
(+) |
|
| L0770 |
conjugate |
|
|
FITC |
| L9895 |
Pseudomonas aeruginosa (PA-I) |
13-13.7 |
— |
— |
gal |
|
|
| L2138 |
Psophocarpus tetragonolobus |
35 |
1 |
— |
galNAc, gal |
|
|
| L3139 |
conjugate |
|
|
Peroxidase |
| L3014 |
conjugate |
|
|
Biotin |
| L3264 |
conjugate |
|
|
FITC |
| Ricinus Communis |
Top of Page |
| L7886 |
Agglutinin, RCA120 |
120 |
4 |
— |
ß-gal |
|
|
| L2390 |
conjugate |
|
|
Agarose |
| L2758 |
conjugate |
|
|
Peroxidase |
| L9514 |
Ricin, A chain |
|
| L4022 |
Ricin, A chain, deglycosylated |
|
| L9639 |
Ricin, B chain |
|
| L6890 |
Sambucus nigra |
|
| L4266 |
Solanum tuberosum |
|
| L9254 |
Tetragonolobus purpureas |
120(A), 58(B), 117(C) |
4;2;4 |
H |
a-L-fuc |
|
|
| L5759 |
conjugate |
|
|
Peroxidase |
| L3134 |
conjugate |
|
|
Biotin |
| L5644 |
conjugate |
|
|
FITC |
| L9640 |
Triticum vulgaris |
36 |
2 |
— |
(glcNAc)2, NeuNAc |
(+) |
|
| L3892 |
conjugate |
|
|
Peroxidase |
| L0390 |
conjugate |
|
|
Peroxidase (inactive) |
| L5142 |
conjugate |
|
|
Biotin |
| L4895 |
conjugate |
|
|
FITC |
| L9884 |
conjugate |
|
|
Evans Blue |
| L1894 |
conjugate |
|
|
Gold, 10nm |
| L1882 |
conjugate |
|
|
Agarose |
| Ulex Europaeus |
Top of Page |
| L5505 |
UEA I |
68 |
— |
H |
a-L-fuc |
|
|
| L8146 |
conjugate |
|
|
Peroxidase |
| L8262 |
conjugate |
|
|
Biotin |
| L9006 |
conjugate |
|
|
FITC |
| L4889 |
conjugate |
|
|
TRITC |
| L6263 |
Vicia faba |
50 |
4(a&ß)a |
— |
man, glc |
(+) |
|
| L4011 |
Vicia villosa |
139 |
4a |
A1+Tn |
galNAc |
|
|
| L9388 |
conjugate |
|
|
Agarose |
| L7513 |
Isolectin B4 |
143 |
4 |
Tn |
galNAc |
|
|
| L7888 |
conjugate |
|
|
Agarose |
| L2662 |
Viscum album |
115g |
4(a&ß)a |
— |
ß-gal |
|
|
| |
Wisteria floribunda |
68 |
2 |
— |
galNAc |
|
|
| L1516 |
conjugate |
|
|
Biotin |
| L2016 |
Reduced |
34 |
1 |
— |
galNAc |
|
|
| L1766 |
conjugate |
|
|
Biotin |
| NOTES: |
|
a. Subunits are of different molecular weights b. MitogeniCfor neuraminidase-treated lymphocytes c. Concentration-dependent mol. wt. change d. Non-agglutinating and mitogeniC |
e. Inhibits mitogeniCactivity of PHA f. Data given for PWM Pa2 g. Data given for VAA(I) h. Agglutinates rabbit, but not human, erythrocytes |
|
back to top Enzyme Explorer
|