| A to C |
| Substrate |
Enzyme |
Product # |
| Acetylthiocholine iodide BioChemika |
Acetylcholinesterase |
01480 |
| 5-Bromo-4-chloro-3-indolyl N-acetyl-b-D-glucosaminide ³98 % |
Acetylglucosaminidase, beta-N- |
B3041 |
| 4-Methylumbelliferyl-7-(6-sulfo-2-acetamido-2-deoxy-b-D-glucopyranoside) sodium salt ~98% (TLC) |
Acetylglucosaminidase, beta-N- |
M0662 |
| 4-Methylumbelliferyl N-acetyl-b-D-glucosaminide |
Acetylglucosaminidase, beta-N- |
M2133 |
| 4-Methylumbelliferyl N-acetyl-a-D-glucosaminide |
Acetylglucosaminidase, beta-N- |
M9881 |
| 4-Nitrophenyl N-acetyl-b-D-glucosaminide 98-100% |
Acetylglucosaminidase, beta-N- |
N9376 |
| 4-Nitrophenyl N-acetyl-b-D-galactosaminide ³98% |
Acetylhexosaminidase, beta-N- |
N9003 |
| sn-Glycerol 3-phosphate bis(cyclohexylammonium) salt BioChemika ³95 % (GC) |
Acid Phosphatase |
49990 |
| Indoxyl acetate BioChemika ³97.0% (TLC) |
Acid Phosphatase |
57420 |
| Naphthol AS-BI phosphate BioChemika for histology ³98.0% (HPLC) |
Acid Phosphatase |
70491 |
| Naphthol AS-BS phosphate BioChemika for histology |
Acid Phosphatase |
70483 |
| Naphthol AS-E phosphate BioChemika for histology |
Acid Phosphatase |
70485 |
| Naphthol AS-MX phosphate BioChemika for histology ³99.0% (HPLC) |
Acid Phosphatase |
70476 |
| Naphthol AS-MX phosphate disodium salt nonahydrate BioChemika for histology ³99.0% (TLC) |
Acid Phosphatase |
70495 |
| Naphthol AS phosphate BioChemika for histology |
Acid Phosphatase |
70471 |
| Naphthol AS-TR phosphate BioChemika for histology ³98.0% (HPLC) |
Acid Phosphatase |
70478 |
| Naphthol AS-TR phosphate disodium salt BioChemika for histology ~4 mg/tablet naphthol AS-TR-phosphate tablets |
Acid Phosphatase |
70467 |
| NBT-BCIP solution BioChemika suitable as substrate for alkaline phosphatase in dot blots |
Acid Phosphatase |
72091 |
| 4-Nitrophenyl phosphate bis(cyclohexylammonium) salt BioChemika for the determination of phosphatase ~95 % (enzymatic) |
Acid Phosphatase |
73736 |
| 4-Nitrophenyl phosphate bis[tris(hydroxymethyl)aminomethane] salt BioChemika for the determination of phosphatase ³97.0 % (enzymatic) |
Acid Phosphatase |
73737 |
| Phenolphthalein bisphosphate pyridine salt BioChemika ³95 % (T) |
Acid Phosphatase |
77630 |
| Thymolphthalein monophosphate disodium salt hydrate BioChemika ~75% (NT) |
Acid Phosphatase |
89362 |
| Tris(4-nitrophenyl) phosphate BioChemika ³95 % (HPLC) |
Acid Phosphatase |
93380 |
| N-Acetyl-Met-Ala ³97 % (HPLC) |
Acylamino acid releasing enzyme |
A1450 |
| Indoxyl acetate BioChemika ³97.0% (TLC) |
Acylase |
57420 |
| Nebularine BioChemika ³98.0 % (HPLC) |
Adenosine deaminase |
82590 |
| N-Benzoyl-Pro-Phe-Arg-p-nitroanilide hydrochloride |
Agkistrodon |
B2133 |
| 3-Fluoro-3-deoxy-D-glucose BioChemika ³99.0 % (TLC) |
Aldolase reductase |
46865 |
| Alkaline Phosphatase Yellow (pNPP) Liquid Substrate System for ELISA liquid |
Alkaline Phosphatase |
A3469 |
| BCIP/NBT-Blue Liquid Substate System for Membranes |
Alkaline Phosphatase |
B3804 |
| BCIP/NBT-Purple Liquid Substate System for Membranes |
Alkaline Phosphatase |
B3679 |
| BCIP/NBT Liquid Substrate System |
Alkaline Phosphatase |
B1911 |
| Bis(4-nitrophenyl) phosphate hydrate purum ³97.0% (dried material, T) |
Alkaline Phosphatase |
15125 |
| SIGMAFAST™ BCIP/NBT tablet |
Alkaline Phosphatase |
B5655 |
| 5-Bromo-4-chloro-3-indolyl phosphate disodium salt |
Alkaline Phosphatase |
B6149 |
| 5-Bromo-4-chloro-3-indolyl phosphate dipotassium salt |
Alkaline Phosphatase |
B6274 |
| 5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt tablet |
Alkaline Phosphatase |
B0274 |
| 5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt ³98% |
Alkaline Phosphatase |
B8503 |
| 5-Bromo-6-chloro-3-indolyl phosphate p-toluidine salt ~97% (HPLC) |
Alkaline Phosphatase |
B5667 |
| 5-Bromo-4-chloro-3-indolyl phosphate disodium salt BioChemika ³98.0 % (HPLC) |
Alkaline Phosphatase |
16668 |
| 5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt BioChemika ³90 % (HPLC) |
Alkaline Phosphatase |
16670 |
| 5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt BioChemika ~25 mg/tablet 5-bromo-4-chloro-3-indolyl phosphate p-toluidine salt tablets |
Alkaline Phosphatase |
16676 |
| 5-Bromo-6-chloro-3-indolyl phosphate p-toluidine salt BioChemika ³95.0 % (HPLC) |
Alkaline Phosphatase |
16672 |
| CDP-Star® Chemiluminescent Substrate 0.25 mM in water |
Alkaline Phosphatase |
C0712 |
| Ammonium cerium(IV) sulfate dihydrate ³99 % |
Alkaline Phosphatase |
31173 |
| 2-Deoxy-a-D-ribose 1-phosphate bis(cyclohexylammonium) salt BioChemika ³95 % (TLC) |
Alkaline Phosphatase |
31173 |
| Fast Red RC tablet 10 mg substrate per tablet |
Alkaline Phosphatase |
F5146 |
| Fast Red TR Salt hemi(zinc chloride) salt |
Alkaline Phosphatase |
F8764 |
| Indoxyl phosphate disodium salt crystalline |
Alkaline Phosphatase |
I5505 |
| Indoxyl phosphate disodium salt BioChemika ³95.0 % (TLC) |
Alkaline Phosphatase |
57440 |
| D-Luciferin 6'-O-phosphate trisodium salt BioChemika ³99.0 % (TLC) |
Alkaline Phosphatase |
62657 |
| D-Luciferin 6'-O-phosphate trisodium salt lyophilized powder |
Alkaline Phosphatase |
L9402 |
| 4-Methylumbelliferyl phosphate BioChemika for fluorescence ³99.0% (HPLC) |
Alkaline Phosphatase |
69607 |
| 4-Methylumbelliferyl phosphate dilithium salt |
Alkaline Phosphatase |
M9044 |
| 4-Methylumbelliferyl phosphate disodium salt |
Alkaline Phosphatase |
M8168 |
| 4-Methylumbelliferyl phosphate disodium salt BioChemika for fluorescence ³99.0% (HPLC) |
Alkaline Phosphatase |
69605 |
| 4-Methylumbelliferyl phosphate |
Alkaline Phosphatase |
M8883 |
| 4-Methylumbelliferyl phosphate (4-MUP) Liquid Substrate System liquid |
Alkaline Phosphatase |
M3168 |
| Naphthol AS-BI phosphate disodium salt |
Alkaline Phosphatase |
N2250 |
| Naphthol AS-BI phosphate BioChemika for histology ³95.0% (HPLC) |
Alkaline Phosphatase |
70482 |
| Naphthol AS-BI phosphate disodium salt BioChemika for histology ³99.0% (TLC) |
Alkaline Phosphatase |
70494 |
| Naphthol AS-E phosphate BioChemika for histology |
Alkaline Phosphatase |
70485 |
| Naphthol AS-MX phosphate |
Alkaline Phosphatase |
N4875 |
| Naphthol AS-MX phosphate BioChemika for histology ³99.0% (HPLC) |
Alkaline Phosphatase |
70476 |
| Naphthol AS-MX phosphate disodium salt nonahydrate BioChemika for histology ³99.0% (TLC) |
Alkaline Phosphatase |
70495 |
| Naphthol AS phosphate BioChemika for histology |
Alkaline Phosphatase |
70471 |
| Naphthol AS-TR phosphate disodium salt |
Alkaline Phosphatase |
N6125 |
| Naphthol AS-TR phosphate BioChemika for histology ³98.0% (HPLC) |
Alkaline Phosphatase |
70478 |
| Naphthol AS-TR phosphate disodium salt BioChemika for histology ~4 mg/tablet naphthol AS-TR-phosphate tablets |
Alkaline Phosphatase |
70467 |
| 1-Naphthyl phosphate sodium salt ³98% (titration) powder |
Alkaline Phosphatase |
N7000 |
| 2-Naphthyl phosphate sodium salt |
Alkaline Phosphatase |
N7375 |
| 1-Naphthyl phosphate disodium salt |
Alkaline Phosphatase |
N7255 |
| 1-Naphthyl phosphate disodium salt BioChemika ³99.0 % (HPLC) |
Alkaline Phosphatase |
71105 |
| 2-Naphthyl phosphate sodium salt BioChemika ³99.0 % (HPLC) |
Alkaline Phosphatase |
71099 |
| 2-Naphthyl phosphate sodium salt BioChemika ³98.0 % (HPLC) |
Alkaline Phosphatase |
71100 |
| 1-Naphthyl phosphate monosodium salt monohydrate BioChemika ³98.0 % (HPLC) |
Alkaline Phosphatase |
71090 |
| NBT-BCIP solution BioChemika suitable as substrate for alkaline phosphatase in dot blots |
Alkaline Phosphatase |
72091 |
| Nitro Blue Tetrazolium tablet |
Alkaline Phosphatase |
N5514 |
| 4-Nitrophenyl phosphate bis(cyclohexylammonium) salt |
Alkaline Phosphatase |
N3129 |
| 4-Nitrophenyl phosphate bis(cyclohexylammonium) salt BioChemika for the determination of phosphatase ~95 % (enzymatic) |
Alkaline Phosphatase |
73736 |
| 4-Nitrophenyl phosphate bis[tris(hydroxymethyl)aminomethane] salt BioChemika for the determination of phosphatase ³97.0 % (enzymatic) |
Alkaline Phosphatase |
73737 |
| 4-Nitrophenyl phosphate di(2-amino-2-ethyl-1,3-propanediol) salt |
Alkaline Phosphatase |
N6260 |
| 4-Nitrophenyl phosphate disodium salt hexahydrate tablet |
Alkaline Phosphatase |
N2640 |
| 4-Nitrophenyl phosphate disodium salt hexahydrate tablet |
Alkaline Phosphatase |
N2765 |
| 4-Nitrophenyl phosphate disodium salt hexahydrate tablet |
Alkaline Phosphatase |
N9389 |
| 4-Nitrophenyl phosphate disodium salt BioChemika purum tablets 20 mg/tablet substrate (remainder binding agent) |
Alkaline Phosphatase |
73724 |
| 4-Nitrophenyl phosphate disodium salt BioChemika ³99.0 % (5 mg substrate per tablet, rest binding agent, TLC) tablets |
Alkaline Phosphatase |
73730 |
| 4-Nitrophenyl phosphate disodium salt hexahydrate BioChemika for enzyme immunoassay ³99.0 % (enzymatic) |
Alkaline Phosphatase |
71768 |
| 4-Nitrophenyl phosphate disodium salt hexahydrate BioChemika ³97.0 % (NT) |
Alkaline Phosphatase |
71770 |
| 4-Nitrophenyl phosphate di(tris) salt |
Alkaline Phosphatase |
N3254 |
| p-Nitrophenyl Phosphate Liquid Substrate System liquid |
Alkaline Phosphatase |
N7653 |
| 4-Nitrophenyl phosphate magnesium salt |
Alkaline Phosphatase |
N2507 |
| 3-Nitrotetrazolium blue chloride BioChemika ³75 % (AT) |
Alkaline Phosphatase |
09972 |
| Nitrotetrazolium blue chloride BioChemika ³88 % (AT) |
Alkaline Phosphatase |
74030 |
| Nitrotetrazolium blue chloride BioChemika ~10 mg/tablet nitrotetrazolium blue chloride |
Alkaline Phosphatase |
74031 |
| Nitrotetrazolium blue chloride BioChemika ³98.0 % (calc. based on dry substance, AT) |
Alkaline Phosphatase |
74032 |
| Phenolphthalein bisphosphate pyridine salt BioChemika ³95 % (T) |
Alkaline Phosphatase |
77630 |
| Phenolphthalein monophosphate bis(cyclohexylammonium) salt BioChemika ³98.0 % (HPLC) |
Alkaline Phosphatase |
77639 |
| Phenolphthalein monophosphate bis(cyclohexylammonium) salt ~90% powder |
Alkaline Phosphatase |
P5758 |
| 3-Phenylumbelliferyl phosphate hemipyridine salt BioChemika for fluorescence ³85% (HPLC) |
Alkaline Phosphatase |
79281 |
| SIGMAFAST™ Fast Red TR/Naphthol AS-MX Tablets tablet To prepare 10 mL |
Alkaline Phosphatase |
F4523 |
| SIGMAFAST™ Fast Red TR/Naphthol AS-MX Tablets tablet To prepare 1 mL |
Alkaline Phosphatase |
F4648 |
| SIGMAFASTä p-Nitrophenyl phosphate Tablets tablet To prepare 5 mL |
Alkaline Phosphatase |
N1891 |
| SIGMAFASTä p-Nitrophenyl phosphate Tablets tablet To prepare 20 mL |
Alkaline Phosphatase |
N2770 |
| 5-(Tetradecanoylamino)fluorescein BioChemika for fluorescence ³95% (HPLC) |
Alkaline Phosphatase |
87180 |
| Tetrazolium Blue Chloride BioChemika indicator for germination, for microbiology ³90% (AT) |
Alkaline Phosphatase |
88190 |
| Thymolphthalein monophosphate disodium salt hydrate BioChemika ~75% (NT) |
Alkaline Phosphatase |
89362 |
| Tris(4-nitrophenyl) phosphate BioChemika ³95 % (HPLC) |
Alkaline Phosphatase |
93380 |
| Na-Benzoyl-DL-arginine 4-nitroanilide hydrochloride BioChemika ³98.0% (AT) |
Amidase |
12920 |
| 3-Amino-2,3-dihydrobenzoic acid hydrochloride purum ³98.0% (calc. based on dry substance, T) |
Aminobutyric acid transaminase, gamma- |
07710 |
| 1-Aminocyclopropanecarboxylic acid BioChemika ³98.0 % (NT) |
Aminocyclopropanecarboxyl oxidase |
07626 |
| Succinyl coenzyme A disodium salt BioChemika ³90 % (HPLC) |
Aminolevulinate synthase, 5- |
85985 |
| 5-Aminolevulinic acid hydrochloride BioChemika ³97.0 % (AT) |
aminolevulinic acid dehydratase, delta- |
08340 |
| L-Alanine 7-amido-4-methylcoumarin trifluoroacetate salt BioChemika for fluorescence ³99.0% (TLC) |
Aminopeptidase |
05198 |
| L-Alanine 4-nitroanilide hydrochloride BioChemika ³99.0 % (TLC) |
Aminopeptidase |
05215 |
| Z-L-arginine 7-amido-4-methylcoumarin hydrochloride BioChemika for fluorescence ³90% (TLC) |
Aminopeptidase |
95938 |
| L-Citrulline 7-amido-4-methylcoumarin hydrobromide BioChemika ³98.0 % (sum of enantiomers, HPLC) |
Aminopeptidase |
27525 |
| Z-Gly-Pro 7-amido-4-methylcoumarin BioChemika for fluorescence ³97.0% (CHN) |
Aminopeptidase |
96280 |
| L-Methionine 7-amido-4-methylcoumarin trifluoroacetate salt BioChemika ³99.0 % (sum of enantiomers, HPLC) |
Aminopeptidase |
64367 |
| L-Proline-2-naphthylamide hydrochloride BioChemika ³99.0 % (TLC) |
Aminopeptidase |
81742 |
| L-Pyroglutamic acid 7-amido-4-methylcoumarin BioChemika ³99.0 % (HPLC) |
Aminopeptidase |
83174 |
| Ammonium thiocyanate puriss. ³99% (AT) |
Aminopeptidase |
11235 |
| L-Aspartic acid b-(7-amido-4-methylcoumarin) BioChemika ³98.0 % (TLC) |
Aminopeptidase |
11235 |
| Fmoc-L-glutamic acid 1-(7-amido-4-methylcoumarin) BioChemika ³98.0 % (HPLC) |
Aminopeptidase A |
02243 |
| L-Glutamic acid 1-(4-nitroanilide) BioChemika ³98.0 % (T) |
Aminopeptidase A |
49622 |
| L-Leucine 7-amido-4-methyl coumarin hydrochloride BioChemika for fluorescence |
Aminopeptidase B |
61888 |
| N-Acetyl-Arg-Gly-Phe-Phe-Pro 4-methoxy-2-naphthylamide >90 % (TLC) solid |
Aminopeptidase M |
C7108 |
| L-Leucine p-nitroanilide BioChemika ³99.0 % (NT) |
Aminopeptidase M |
61910 |
| L-Phenylalanine 7-amido-4-methylcoumarin trifluoroacetate salt BioChemika for fluorescence ³99.0% (sum of enantiomers, HPLC) |
Aminopeptidase M |
78089 |
| 4-Nitrophenyl a-D-maltopentaoside ³98% |
Amylase |
N1519 |
| Starch Azure Insoluble corn starch covalently linked with Remazol Brilliant Blue R |
Amylase |
S7776 |
| Starch Azure Potato starch covalently linked with Remazol Brilliant Blue R |
Amylase |
S7629 |
| Amylopectin Azure |
Amylase, alpha- |
A4640 |
| Amylose–Remazol Brilliant Blue R |
Amylase, alpha- |
A3508 |
| Amylose–Remazol Brilliant Blue R BioChemika |
Amylase, alpha- |
10140 |
| 2-Chloro-4-nitrophenyl-b-D-maltoheptaoside BioChemika ~80 % (HPLC) |
Amylase, alpha- |
25590 |
| 2-Chloro-4-nitrophenyl-b-D-maltotrioside BioChemika ³96.0 % (HPLC) |
Amylase, alpha- |
25591 |
| 4-Nitrophenyl a-D-maltoside |
Amylase, alpha- |
N5885 |
| Starch dyed with Remazol brilliant blue R BioChemika |
Amylase, alpha- |
85641 |
| Z-Val-Lys-Met 7-amido-4-methylcoumarin |
Amyloid A4-generating enzymes |
C2681 |
| N-[3-(2-Furyl)acryloyl]-Phe-Gly-Gly |
Angiotensin Converting Enzyme |
F7131 |
| Hippuryl-Gly-Gly |
Angiotensin Converting Enzyme |
H1763 |
| Hippuryl-His-Leu acetate salt |
Angiotensin Converting Enzyme |
H4884 |
| N-Hippuryl-His-Leu tetrahydrate |
Angiotensin Converting Enzyme |
H1635 |
| N-Hippuryl-His-Leu tetrahydrate BioChemika ³99.0 % (HPLC) |
Angiotensin-converting enzyme |
53285 |
| 4-Nitrophenyl a-L-arabinofuranoside BioChemika ³99.0 % (TLC) |
Arabinofuranosidase III |
73616 |
| 4-Nitrophenyl a-L-arabinofuranoside ³98% (TLC) |
Arabinofuranosidase III, alpha-L- |
N3641 |
| 4-Methylumbelliferyl a-L-arabinopyranoside |
Arabinosidase |
M3167 |
| 5-Hydroxy-L-tryptophan solid |
Aromatic amino acid decarboxylase, L- |
H9772 |
| Tryptamine hydrochloride BioChemika ³98.0 % (AT) |
Arylamine N-methyltransferase |
93650 |
| 6-Chloro-3-indolyl phosphate p-toluidine salt BioChemika ³98.0 % (HPLC) |
Aryl-sulfatase |
24908 |
| 4-Nitrocatechol sulfate dipotassium salt monohydrate BioChemika ³97.0 % (T) |
Arylsulfatases A and B |
56020 |
| Na-Acetyl-L-asparagine BioChemika ³98.0 % (T) |
Asparaginase II |
00915 |
| Oxaloacetic acid BioChemika ³98.0 % (T) |
Aspartate transaminase |
75660 |
| Adenosine 5'-triphosphate P3-[1-(2-nitrophenyl)ethyl ester] disodium salt BioChemika ³95% (HPLC) |
ATPase |
02070 |
| N-Methoxysuccinyl-Phe-Leu-Phe-7-amido-4-trifluoromethylcoumarin solid |
ATP-dependent protease |
M4813 |
| N-Methoxysuccinyl-Phe-Leu-Phe 4-methoxy-2-naphthylamide ³90 % solid |
ATP-dependent protease |
M4938 |
| Amyloid b/A4 Precursor Protein 770 Fragment 667-676 ³96 % (HPLC) solid |
beta-Secretase |
A3222 |
| [Asn670, Leu671]-Amyloid b/A4 Precursor Protein 770 Fragment 667-676 trifluoroacetate salt ³95 % (HPLC) solid |
beta-Secretase |
A1972 |
| DABCYL-[Asn670, Leu671]-Amyloid b/A4 Precursor Protein 770 fragment 661-675-EDANS trifluoroacetate salt ³97 % (HPLC) solid |
beta-Secretase |
A4972 |
| DABCYL-[Asn670, Leu671]-Amyloid b/A4 Precursor Protein 770 Fragment 669-674-EDANS ammonium salt ³95 % (HPLC) solid |
beta-Secretase |
A5097 |
| 7-Methoxycoumarin-4-acetyl-[Asn670, Leu671]-Amyloid b/A4 Precursor Protein 770 Fragment 669-674-(2,4-dinitrophenyl)Lys trifluoroacetate salt ³96 % (HPLC) |
beta-Secretase |
A1597 |
| 7-Methoxycoumarin-4-acetyl-[Asn670, Leu671]-Amyloid b/A4 Precursor Protein 770 Fragment 667-674-(2,4-dinitrophenol)Dap ³97 % (HPLC) |
beta-Secretase |
A1222 |
| 7-Methoxycoumarin-4-acetyl-[Asn670, Leu671]-Amyloid b/A4 Precursor Protein 770 Fragment 667-676-(2,4-dinitrophenyl)Lys-Arg-Arg amide ³90 % (HPLC) |
beta-Secretase |
A1472 |
| 7-Methoxycoumarin-4-acetyl-[Asn670,Leu671]-Amyloid b/A4 Precursor Protein 770 (667-676)-(2,4-dinitrophenyl)Lys-Arg-Arg Amide ³96 % (HPLC) |
beta-Secretase |
A1347 |
| [Val671]-Amyloid b/A4 Precursor Protein 770 Fragment 667-676 trifluoroacetate salt ³95 % (HPLC) solid |
beta-Secretase |
A2722 |
| N-(+)-Biotinyl-4-aminobenzoic acid BioChemika ³98.0 % (HPLC) |
Biotinidase |
14408 |
| Zinc puriss. ³99.99% granulated |
Biotinidase |
14404 |
| Biotin 4-amidobenzoic acid sodium salt BioChemika ³99.0 % (TLC) |
Biotinidase |
14404 |
| Zinc purum ³99% powder |
Biotinidase |
14409 |
| Biocytin BioChemika ³97.0 % (HPLC) |
Biotinidase |
14409 |
| Na-Benzoyl-L-arginine 4-nitroanilide hydrochloride BioChemika ³98.0 % (enzymatic) |
Bromelain |
12915 |
| Z-L-Lys-ONp hydrochloride |
Bromelain |
C3637 |
| Z-Lys-ONp hydrochloride purum ³98.0% (AT) |
Bromelain |
96895 |
| 4-Methylumbelliferyl butyrate BioChemika for fluorescence ³95% (HPCE) |
Butyrate esterase |
19362 |
| 4-Nitrophenyl butyrate BioChemika ³98.0 % (HPLC) |
Butyrylcholinesterase |
19368 |
| Autocamtide 2 trifluoroacetate salt ³97 % (HPLC) |
Calmodulin-dependent protein kinase II |
A0591 |
| Syntide 2 ³97% (HPLC) |
Calmodulin-dependent protein kinase II |
S2525 |
| Syntide 2 BioChemika ³99.0 % (HPLC) |
Calmodulin-dependent protein kinase II |
86221 |
| Calcium/calmodulin-dependent protein kinase II-d fragment 345-358 |
Calmodulin-dependent protein kinase IV |
C0843 |
| L-Methionine 7-amido-4-methylcoumarin trifluoroacetate salt BioChemika ³99.0 % (sum of enantiomers, HPLC) |
Calpain |
64367 |
| N-Succinyl-Leu-Leu-Val-Tyr-7-Amido-4-Methylcoumarin ³90% (HPLC) |
Calpain |
S6510 |
| N-Succinyl-Leu-Tyr-7-amido-4-methylcoumarin |
Calpain |
S1153 |
| Acetyl-L-phenylalanyl-L-3-thiaphenylalanine BioChemika ³98.0 % (HPLC) |
Carboxypeptidase A |
29223 |
| Hippuryl-Lys |
Carboxypeptidase B |
H6750 |
| Acetyl-Lys-D-Ala-D-Ala ³95 % (HPLC) |
Carboxypeptidase G |
A6950 |
| N-(3-[2-Furyl]acryloyl)-Ala-Lys powder |
Carboxypeptidase N |
F5882 |
| Na,Ne-Diacetyl-Lys-D-Ala-D-Ala |
Carboxypeptidase, penicillin-sensitive D-alanine |
D9904 |
| Na,Ne-Diacetyl-Lys-D-Ala-D-lactic acid |
Carboxypeptidase, penicillin-sensitive D-alanine |
D2279 |
| N-(3-[2-Furyl]acryloyl)-Phe-Phe |
Carboxypeptidase, Serum |
F7133 |
| Hippuryl-L-phenylalanine BioChemika ³97.0 % (TLC) |
CarboxypeptidaseA |
53284 |
| Hippuryl-L-phenylalanine |
CarboxypeptidaseA |
H6875 |
| L-Carnosine BioChemika ³99.0 % (NT) |
Carnosinase |
22030 |
| Casein Kinase II substrate ³97% (HPLC) |
Casein Kinase II |
C2460 |
| N-Acetyl-Val-Ala-Asp-7-amido-4-trifluoromethylcoumarin >98 % (HPLC) solid |
Caspase (general) |
A0723 |
| N-Acetyl-Val-Ala-Asp p-nitroanilide >98 % (HPLC) solid |
Caspase (general) |
A0848 |
| L-Asp-Asp-Rhodamine trifluoroacetate salt >98 % solid |
Caspase (general) |
D5189 |
| N-Acetyl-Trp-Glu-His-Asp-7-amido-4-trifluoromethylcoumarin trifluoroacetate salt ³97 % (HPLC) powder |
Caspase 1 |
A6720 |
| N-Acetyl-Trp-Glu-His-Asp-p-nitroanilide ³98 % (HPLC) powder |
Caspase 1 |
A6845 |
| N-Acetyl-Tyr-Val-Ala-Asp p-nitroanilide ³97% (HPLC) powder |
Caspase 1 |
A3831 |
| N-Acetyl-Trp-Glu-His-Asp-7-amido-4-methylcoumarin ³97 % (HPLC) powder |
Caspase 1 |
A0216 |
| N-Acetyl-Tyr-Val-Ala-Asp-7-amido-4-trifluoromethylcoumarin ³95 % (HPLC) powder |
Caspase 1 |
A9965 |
| 7-Methoxycoumarin-4-acetyl-Tyr-Val-Ala-Asp-Ala-Pro-(2,4-dinitrophenyl)Lys ³97 % (HPLC) |
Caspase 1 |
M5295 |
| N-Acetyl-Tyr-Val-Ala-Asp-7-amido-4-methylcoumarin ³99% (TLC) powder |
Caspase 1 |
A2452 |
| N-Acetyl-Val-Asp-Val-Ala-Asp-7-amido-4-trifluoromethylcoumarin ³97 % (HPLC) powder |
Caspase 2 |
A5345 |
| N-Acetyl-Val-Asp-Val-Ala-Asp-p-nitroanilide ³97 % (HPLC) powder |
Caspase 2 |
A5470 |
| N-Acetyl-Val-Asp-Val-Ala-Asp-7-amido-4-trifluoromethylcoumarin ³97 % (HPLC) powder |
Caspase 2 |
A5345 |
| N-Acetyl-Val-Asp-Val-Ala-Asp-p-nitroanilide ³97 % (HPLC) powder |
Caspase 2 |
A5470 |
| N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-trifluoromethylcoumarin ³90 % (HPLC) powder |
Caspase 3 |
A0466 |
| N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-trifluoromethylcoumarin ³90 % (HPLC) powder |
Caspase 3 |
A0466 |
| N-Acetyl-Asp-Gln-Met-Asp-7-amido-4-trifluoromethylcoumarin ³90 % (HPLC) powder |
Caspase 3 |
A5595 |
| N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-methylcoumarin ³97% (HPLC) powder |
Caspase 3 |
A1086 |
| N-Acetyl-Asp-Glu-Val-Asp p-nitroanilide ³97% (HPLC) powder |
Caspase 3 |
A2559 |
| N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-trifluoromethylcoumarin ³90 % (HPLC) powder |
Caspase 3 |
A0466 |
| N-Acetyl-Asp-Glu-Val-Asp p-nitroanilide ³97% (HPLC) powder |
Caspase 3 |
A2559 |
| DFF45/ICAD human >85 % (SDS-PAGE) recombinant, expressed in Escherichia coli solution |
Caspase 3 |
D3815 |
| N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-methylcoumarin ³97% (HPLC) powder |
Caspase 3 |
A1086 |
| N-Acetyl-Asp-Glu-Val-Asp p-nitroanilide ³97% (HPLC) powder |
Caspase 3 |
A2559 |
| N-Acetyl-Asp-Gln-Met-Asp-7-amido-4-trifluoromethylcoumarin ³90 % (HPLC) powder |
Caspase 3 |
A5595 |
| N-Acetyl-Leu-Glu-Val-Asp-7-amido-4-trifluoromethylcoumarin ³95 % (HPLC) solid |
Caspase 4 |
A2099 |
| N-Acetyl-Tyr-Val-Ala-Asp p-nitroanilide ³97% (HPLC) powder |
Caspase 4 |
A3831 |
| N-Acetyl-Tyr-Val-Ala-Asp-7-amido-4-methylcoumarin ³99% (TLC) powder |
Caspase 4 |
A2452 |
| N-Acetyl-Tyr-Val-Ala-Asp-7-amido-4-trifluoromethylcoumarin ³95 % (HPLC) powder |
Caspase 4 |
A9965 |
| N-Acetyl-Trp-Glu-His-Asp-7-amido-4-trifluoromethylcoumarin trifluoroacetate salt ³97 % (HPLC) powder |
Caspase 5 |
A6720 |
| N-Acetyl-Trp-Glu-His-Asp-p-nitroanilide ³98 % (HPLC) powder |
Caspase 5 |
A6845 |
| N-Acetyl-Trp-Glu-His-Asp-p-nitroanilide ³98 % (HPLC) powder |
Caspase 5 |
A6845 |
| N-Acetyl-Trp-Glu-His-Asp-p-nitroanilide ³98 % (HPLC) powder |
Caspase 5 |
A6845 |
| N-Acetyl-Val-Glu-His-Asp-7-amido-4-trifluoromethylcoumarin trifluoroacetate salt ³98 % (HPLC) solid |
Caspase 6 |
A2097 |
| N-Acetyl-Val-Glu-Ile-Asp-7-amido-4-methylcoumarin ³90 % (HPLC) powder |
Caspase 6 |
A0341 |
| N-Acetyl-Val-Glu-Ile-Asp-7-amido-4-trifluoromethylcoumarin ³97 % (HPLC) powder |
Caspase 6 |
A5095 |
| N-Acetyl-Val-Glu-Ile-Asp-7-amido-4-methylcoumarin ³90 % (HPLC) powder |
Caspase 6 |
A0341 |
| N-Acetyl-Val-Glu-Ile-Asp-p-nitroanilide ³97 % (HPLC) powder |
Caspase 6 |
A5220 |
| CASPASE-6 ASSAY KIT, FLUORIMETRIC |
Caspase 6 |
CASP-6-F |
| N-Acetyl-Val-Glu-Ile-Asp-7-amido-4-methylcoumarin ³90 % (HPLC) powder |
Caspase 6 |
A0341 |
| N-Acetyl-Val-Glu-Ile-Asp-7-amido-4-trifluoromethylcoumarin ³97 % (HPLC) powder |
Caspase 6 |
A5095 |
| N-Acetyl-Val-Glu-Ile-Asp-p-nitroanilide ³97 % (HPLC) powder |
Caspase 6 |
A5220 |
| N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-trifluoromethylcoumarin ³90 % (HPLC) powder |
Caspase 7 |
A0466 |
| N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-trifluoromethylcoumarin ³90 % (HPLC) powder |
Caspase 7 |
A0466 |
| N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-methylcoumarin ³97% (HPLC) powder |
Caspase 7 |
A1086 |
| N-Acetyl-Asp-Glu-Val-Asp p-nitroanilide ³97% (HPLC) powder |
Caspase 7 |
A2559 |
| N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-methylcoumarin ³97% (HPLC) powder |
Caspase 7 |
A1086 |
| N-Acetyl-Asp-Glu-Val-Asp p-nitroanilide ³97% (HPLC) powder |
Caspase 7 |
A2559 |
| N-Acetyl-Ala-Glu-Val-Asp-7-amido-4-trifluoromethylcoumarin >96 % (HPLC) solid |
Caspase 8 |
A0973 |
| N-Acetyl-Ala-Glu-Val-Asp-7-amido-4-trifluoromethylcoumarin >96 % (HPLC) solid |
Caspase 8 |
A0973 |
| N-Acetyl-Ile-Glu-Thr-Asp-p-nitroanilide ³95 % |
Caspase 8 |
A9968 |
| N-Acetyl-Leu-Glu-Thr-Asp-7-Amido-4-trifluoromethylcoumarin ³97 % (HPLC) powder |
Caspase 8 |
A6095 |
| N-Acetyl-Ile-Glu-Thr-Asp-7-Amido-4-methylcoumarin ³97 % (HPLC) powder |
Caspase 8 |
A4188 |
| CASPASE-8 ASSAY KIT, COLORIMETRIC |
Caspase 8 |
CASP-8-C |
| CASPASE-8 ASSAY KIT, FLUORIMETRIC |
Caspase 8 |
CASP-8-F |
| Z-Ile-Glu-Thr-Asp 7-amido-4-trifluoromethylcoumarin ³95 % (HPLC) |
Caspase 8 |
C5599 |
| N-Acetyl-Ile-Glu-Thr-Asp-7-Amido-4-methylcoumarin ³97 % (HPLC) powder |
Caspase 8 |
A4188 |
| N-Acetyl-Leu-Glu-Thr-Asp-7-Amido-4-trifluoromethylcoumarin ³97 % (HPLC) powder |
Caspase 8 |
A6095 |
| N-Acetyl-Ile-Glu-Thr-Asp-p-nitroanilide ³95 % |
Caspase 8 |
A9968 |
| Z-Ile-Glu-Thr-Asp 7-amido-4-trifluoromethylcoumarin ³95 % (HPLC) |
Caspase 8 |
C5599 |
| N-Acetyl-Leu-Glu-His-Asp-7-amido-4-trifluoromethylcoumarin ³95 % (HPLC) powder |
Caspase 9 |
A5845 |
| N-Acetyl-Val-Glu-His-Asp-7-amido-4-trifluoromethylcoumarin trifluoroacetate salt ³98 % (HPLC) solid |
Caspase 9 |
A2097 |
| N-Acetyl-Leu-Glu-His-Asp-7-amido-4-trifluoromethylcoumarin ³95 % (HPLC) powder |
Caspase 9 |
A5845 |
| N-Acetyl-Leu-Glu-His-Asp-p-nitroanilide ³95 % (HPLC) powder |
Caspase 9 |
A5970 |
| Hydrogen peroxide solution puriss. p.a. ACS reagent ³30% (RT) |
Catalase |
95300 |
| Z-Phe-Ala puriss. ³99.0% (T) |
Cathepsin A |
97029 |
| Z-Ala-Arg-Arg 4-methoxy-b-naphthylamide acetate salt |
Cathepsin B |
C8536 |
| Z-Phe-Arg 4-methoxy-b-naphthylamide hydrochloride |
Cathepsin B |
C3282 |
| Z-Val-Lys-Lys-Arg-4-methoxy-b-naphthylamide trihydrochloride |
Cathepsin B |
C2772 |
| Hippuryl-Arg |
Cathepsin B |
H2508 |
| Na-Benzoyl-L-arginine 4-methoxy-b-naphthylamide hydrochloride |
Cathepsin B |
B0885 |
| Z-Arg-Arg-7-amido-4-methylcoumarin hydrochloride |
Cathepsin B |
C5429 |
| Z-Arg-Arg 4-methoxy-b-naphthylamide acetate salt |
Cathepsin B |
C5520 |
| N-Succinyl-Leu-Leu-Val-Tyr-7-Amido-4-Methylcoumarin ³90% (HPLC) |
Cathepsin B |
S6510 |
| Gly-Phe b-naphthylamide crystalline |
Cathepsin C |
G4255 |
| Gly-Phe b-naphthylamide crystalline |
Cathepsin C |
G9512 |
| Gly-Phe p-nitroanilide |
Cathepsin C |
G0142 |
| Ser-Tyr b-naphthylamide powder |
Cathepsin C |
S5628 |
| N-Acetyl-Arg-Gly-Phe-Phe-Pro-7-amido-4-methylcoumarin solid |
Cathepsin D |
C6358 |
| N-Acetyl-Arg-Gly-Phe-Phe-Pro-7-amido-4-trifluoromethylcoumarin >95 % (HPLC) solid |
Cathepsin D |
C6983 |
| N-Acetyl-Arg-Gly-Phe-Phe-Pro 4-methoxy-2-naphthylamide >90 % (TLC) solid |
Cathepsin D |
C7108 |
| Boc-Phe-Ala-Ala-p-Nitro-Phe-Phe-Val-Leu 4-hydroxymethylpyridine ester |
Cathepsin D |
B1644 |
| 7-Methoxycoumarin-4-Acetyl-Gly-Lys-Pro-Ile-Leu-Phe-Phe-Arg-Leu-Lys(DNP)-D- Arg-amide >97 % (HPLC) solid |
Cathepsin D |
M0938 |
| 7-Methoxycoumarin-4-Acetyl-Gly-Lys-Pro-Ile-Leu-Phe-Phe-Arg-Leu-Lys(DNP)-D- Arg-amide >97 % (HPLC) solid |
Cathepsin E |
M0938 |
| N-Methoxysuccinyl-Ala-Ala-Pro-Met p-nitroanilide >95 % (HPLC) |
Cathepsin G |
M7771 |
| N-Succinyl-Ala-Ala-Pro-Phe p-nitroanilide |
Cathepsin G |
S7388 |
| N-Succinyl-Ala-Ala-Pro-Phe thiobenzyl ester |
Cathepsin G |
S6518 |
| N-Succinyl-Gly-Gly-Phe-7-amido-4-trifluoromethylcoumarin ³98 % (TLC) solid |
Cathepsin G |
S4689 |
| N-Succinyl-Gly-Gly-Phe-4-methoxy-2-naphthylamide ³98 % (TLC) solid |
Cathepsin G |
S4814 |
| N-Succinyl-Phe-Leu-Phe thiobenzyl ester |
Cathepsin G |
S6643 |
| Z-Phe-Arg 4-methoxy-b-naphthylamide hydrochloride |
Cathepsin L |
C3282 |
| N-Succinyl-Ala-Ala-Pro-Phe p-nitroanilide |
Cathepsin, Leukocyte |
S7388 |
| Cellotetraose BioChemika ~95 % (HPLC) |
Cellulase |
22164 |
| Cellulose Azure |
Cellulase |
C1052 |
| 2-Chloro-4-nitrophenyl-b-D-cellobioside BioChemika chromophoric enzyme substrate ³98.0 % (HPCE) |
Cellulase |
25586 |
| 4-Methylumbelliferyl b-D-cellobioside |
Cellulase |
M6018 |
| 2-Nitrophenyl b-D-cellobioside |
Cellulase |
N4764 |
| 4-Nitrophenyl b-D-cellobioside |
Cellulase |
N5759 |
| 4-Nitrophenyl b-D-cellobioside BioChemika ³99.0 % (HPLC) |
Cellulase |
73617 |
| Chitin azure |
Chitinase |
C3020 |
| 4-Methylumbelliferyl-N,N'-diacetyl-b-D-chitobioside monohydrate BioChemika for fluorescence ³99.0% (HPLC) |
Chitinase |
69589 |
| 4-Nitrophenyl N,N'-diacetyl-b-D-chitobioside |
Chitinase |
N6133 |
| 4-Nitrophenyl N,N'-diacetyl-b-D-chitobioside BioChemika ³99.0 % (TLC) |
Chitinase |
73623 |
| CHKtide, CHK1 Substrate Peptide liquid |
CHK1 |
C7734 |
| 2-Chloro-5,5-dimethyl-1,3-cyclohexanedione BioChemika suitable as substrate for chloroperoxidase ³98.0 % (AT) |
Chloroperoxidase |
24368 |
| 7-Acetoxy-1-methylquinolinium iodide BioChemika for fluorescence ³98.0% (HPCE) |
Cholinesterase |
00871 |
| Benzoylcholine chloride BioChemika ³98.0 % (AT) |
Cholinesterase |
12950 |
| Butyrylcholine iodide BioChemika ³99.0 % (AT) |
Cholinesterase |
20780 |
| b-Carbonaphthoxycholine iodide |
Cholinesterase |
C7125 |
| 2,6-Dichlorophenolindophenyl acetate BioChemika ³97.0% (CHN) |
Cholinesterase |
36190 |
| Pencycuron PESTANAL®, analytical standard |
Cholinesterase |
36190 |
| Indoxyl acetate BioChemika ³97.0% (TLC) |
Cholinesterase |
57420 |
| 4-Nitrophenyl butyrate |
Cholinesterase |
|
| Resorufin butyrate BioChemika for fluorescence ³95% (TLC) |
Cholinesterase |
83637 |
| Succinylcholine chloride dihydrate BioChemika ³99.0% (AT) |
Cholinesterase |
85980 |
| N-Succinyl-Ala-Ala-Pro-Phe p-nitroanilide |
Chymotrypsin, alpha |
S7388 |
| N-Succinyl-Ala-Ala-Pro-Phe-7-amido-4-methylcoumarin |
Chymotrypsin, alpha |
S9761 |
| N-Acetyl-L-tyrosine ethyl ester monohydrate BioChemika ³99.0 % (sum of enantiomers, HPLC) |
Chymotrypsin, alpha- |
01530 |
| Ala-Ala-Phe-7-amido-4-methylcoumarin |
Chymotrypsin, alpha- |
A3401 |
| Ala-Ala-Val-Ala p-nitroanilide |
Chymotrypsin, alpha- |
A9273 |
| N-Benzoyl-L-tyrosine ethyl ester BioChemika ³98.0% (NT) |
Chymotrypsin, alpha- |
13110 |
| Magnesium according to Grignard ³99.5 % turnings |
Chymotrypsin, alpha- |
13110 |
| N-Benzoyl-L-tyrosine ethyl ester |
Chymotrypsin, alpha- |
B6125 |
| N-Benzoyl-L-tyrosine p-nitroanilide |
Chymotrypsin, alpha- |
B6760 |
| N-Benzoyl-L-tyrosine p-amidobenzoic acid sodium salt BioChemika lyophilized ³98.0 % (sum of enantiomers, HPLC) |
Chymotrypsin, alpha- |
13135 |
| Glutaryl-Ala-Ala-Phe-4-methoxy-b-naphthylamide |
Chymotrypsin, alpha- |
G3769 |
| N-Glutaryl-Gly-Gly-Phe-b-naphthylamide |
Chymotrypsin, alpha- |
G4636 |
| Glutaryl-L-phenylalanine 7-amido-4-methylcoumarin BioChemika for fluorescence ³98.0% (HPLC) |
Chymotrypsin, alpha- |
49737 |
| N-Glutaryl-L-phenylalanine p-nitroanilide BioChemika ³98.0 % (HPLC) |
Chymotrypsin, alpha- |
49738 |
| N-(N-Glutaryl-L-phenylalanyl)-2-aminoacridone BioChemika for fluorescence ³98.0% (HPLC) |
Chymotrypsin, alpha- |
49742 |
| 4-Methylumbelliferyl p-trimethylammoniocinnamate chloride powder |
Chymotrypsin, alpha- |
M4507 |
| N-Succinyl-Ala-Ala-Pro-Phe 7-amido-4-trifluoromethylcoumarin ³99 % (TLC) solid |
Chymotrypsin, alpha- |
S5189 |
| N-Succinyl-Ala-Ala-Pro-Phe 4-methoxy-2-naphthylamide ³98 % (TLC) solid |
Chymotrypsin, alpha- |
S5314 |
| N-Succinyl-Ala-Ala-Pro-Phe p-nitroanilide BioChemika ³99.0 % (HPLC) |
Chymotrypsin, alpha- |
85977 |
| N-Succinyl-Gly-Gly-Phe-p-nitroanilide |
Chymotrypsin, alpha- |
S1899 |
| N-Succinyl-L-phenylalanine-p-nitroanilide |
Chymotrypsin, alpha- |
S2628 |
| N-(N-Succinyl-L-phenylalanyl)-2-aminoacridone BioChemika for fluorescence ³98.0% (HPCE) |
Chymotrypsin, alpha- |
85992 |
| Oxaloacetic acid BioChemika ³98.0 % (T) |
Citrate synthase |
75660 |
| c-Jun human 40-50 % (SDS-PAGE) recombinant, expressed in Escherichia coli truncated human c-Jun sequence-GST fusion protein 1-169-GST, soluble |
c-Jun Kinase |
C5859 |
| c-Jun Kinase substrate from rat >90 % (SDS-PAGE) recombinant, expressed in Escherichia coli buffered aqueous solution |
c-Jun Kinase |
C3233 |
| Succinyl coenzyme A disodium salt BioChemika ³90 % (HPLC) |
CoA ligase, succinate- |
85985 |
| N-[3-(2-Furyl)acryloyl]-Leu-Gly-Pro-Ala |
Collagenase |
F5135 |
| N-(2,4-Dinitrophenyl)-Pro-Leu-Gly-Cys(Me)-His-Ala-D-Arg amide ³95 % (HPLC) |
Collagenase |
D0296 |
| N-[3-(2-Furyl)acryloyl]-Leu-Gly-Pro-Ala BioChemika ³99.0% (HPLC) |
Collagenase |
48173 |
| Z-Gly-Pro-Gly-Gly-Pro-Ala |
Collagenase |
C8008 |
| H-Lys(Z)-OH 98 % |
Collagenase |
C8008 |
| Z-Gly-Pro-Leu-Gly-Pro |
Collagenase |
C0627 |
| COLLAGEN-FLUORESCEIN |
Collagenase |
C4486 |
| Collagen Type I-FITC Conjugate bovine skin ~1 mg/mL solution |
Collagenase |
C4361 |
| Collagen-Fluorescein bovine |
Collagenase |
C5858 |
| Collagenase Chromophore-Substrate (for quantitative Collagenase-Determination) BioChemika |
Collagenase |
27668 |
| Collagenase Chromophore-Substrate (for quantitative Collagenase-Determination) Components A + B BioChemika |
Collagenase |
27669 |
| Collagenase Chromophore-Substrate (for quantitative Collagenase-Determination) Components A + B BioChemika |
Collagenase |
27671 |
| Collagenase-Substrate (for quantitative Collagenase-Determination) Components A + B BioChemika |
Collagenase |
27670 |
| Collagenase-Substrate (for quantitative Collagenase-Determination) Components A + B BioChemika |
Collagenase |
27672 |
| Collagenase-Substrate (for quantitative Collagenase-Determination) BioChemika |
Collagenase |
27673 |
| Collagenase-Substrate (for quantitative Collagenase-Determination) Component B BioChemika |
Collagenase |
27674 |
| Glycolic acid solution ~57 % |
Collagenase |
27674 |
| 7-Methoxycoumarin-4-acetyl-Pro-(cyclohexyl)-Ala-Gly-Nva-His-Ala-L-(2,4-dinitrop henyl)diaminoproponic acid solid |
Collagenase-3 |
M3314 |
| Boc-Leu-Gly-Arg-7-amido-4-methylcoumarin acetate salt |
Convertase |
B4511 |
| Urea BioChemika Ultra ³99.5% (T) |
Cyanamide hydratase |
51458 |
| Resorufin benzyl ether |
Cytochrome P450 |
B1532 |
| Resorufin methyl ether |
Cytochrome P450 |
M1544 |
| Resorufin ethyl ether |
Cytochrome P450 |
E3763 |
| 7-Benzyloxy-4-trifluoromethylcoumarin 99 % (by 1H-NMR) solid |
Cytochrome P450 1A2 |
B5057 |
| 7-Ethoxycoumarin BioChemika for fluorescence ³99.0% (HPLC) |
Cytochrome P450 2B6 |
02644 |
| 7-Methoxy-4-trifluoromethylcoumarin 99 % (by 1H-NMR) |
Cytochrome P450 2C9 |
M8189 |
| 7-Benzyloxyquinoline 99% (by 1H-NMR) solid |
Cytochrome P450 3A4 |
B5182 |
| 7-Benzyloxy-4-trifluoromethylcoumarin 99 % (by 1H-NMR) solid |
Cytochrome P450 3A4 |
B5057 |
| 3-Nitrotetrazolium blue chloride BioChemika ³75 % (AT) |
Cytokinin dehydrogenase |
09972 |
| Nitrotetrazolium blue chloride BioChemika ³88 % (AT) |
Cytokinin dehydrogenase |
74030 |
| Nitrotetrazolium blue chloride BioChemika ~10 mg/tablet nitrotetrazolium blue chloride |
Cytokinin dehydrogenase |
74031 |
| Nitrotetrazolium blue chloride BioChemika ³98.0 % (calc. based on dry substance, AT) |
Cytokinin dehydrogenase |
74032 |
| Tetrazolium Blue Chloride BioChemika indicator for germination, for microbiology ³90% (AT) |
Cytokinin dehydrogenase |
88190 |
| N-Acetyl-Gly(t-butyl)-Gly(t-butyl)-Asn(Methyl)2-Ala-7-amido-4-methylcoumari n ³97 % (HPLC) solid |
Cytomegalovirus (HCMV) protease |
A2347 |
| 2-Aminobenzoyl-Gly(t-butyl)-Asn(methyl)2-Ala-Ser-Ser-Arg-Leu-Tyr(NO2)-Tyr-Arg trifluoroacetate salt >94 % (HPLC) |
Cytomegalovirus (HCMV) protease |
A9846 |
| Resorufin acetate BioChemika for fluorescence ³98.0% (TLC) |
Cytosolic Aldehyde Dehydrogenase (Esterase Activity) |
83636 |
| D to N |
Back to top |
| Substrate |
Enzyme |
Product # |
| Resorufin pentyl ether BioChemika for fluorescence ³98.0% (HPLC) |
Dealkylase |
77105 |
| Neotetrazolium chloride form BioChemika for microbiology for the detection of reducing fermentation systems ³75% (T) |
Dehydrogenase |
72150 |
| Thiazolyl blue formazan BioChemika |
Dehydrogenase |
88417 |
| Thiazolyl Blue Tetrazolium Bromide BioChemika ³97.0 % (enzymatic) |
Dehydrogenase |
88415 |
| 2,3,5-Triphenyltetrazolium chloride BioChemika ³99.0 % (AT) |
Dehydrogenase |
93140 |
| 1,3,5-Triphenyltetrazolium formazan BioChemika ³97.0 % (UV) |
Dehydrogenase |
93145 |
| N-Acetyl-L-phenylalanyl-3,5-diiodo-L-tyrosine BioChemika ³97.0 % (CHN) |
Diiodotyrosine transaminase |
01408 |
| Lys-Ala-7-amido-4-methylcoumarin dihydrochloride |
Dipeptidyl aminopeptidase |
L8139 |
| Gly-Arg 4-methoxy-b-naphthylamide dihydrochloride |
Dipeptidyl aminopeptidase I |
G1013 |
| Lys-Ala 4-methoxy-b-naphthylamide dihydrochloride |
Dipeptidyl aminopeptidase II |
L2270 |
| Arg-Arg b-naphthylamide trihydrochloride |
Dipeptidyl aminopeptidase III |
A3261 |
| Arg-Arg 4-methoxy-b-naphthylamide trihydrochloride |
Dipeptidyl aminopeptidase III |
A9398 |
| Arg-Pro p-nitroanilide acetate salt |
Dipeptidyl aminopeptidase IV |
A1204 |
| Gly-Pro 4-methoxy-b-naphthylamide hydrochloride powder |
Dipeptidyl aminopeptidase IV |
G9262 |
| Gly-Pro 4-methoxy-b-naphthylamide |
Dipeptidyl aminopeptidase IV |
G9137 |
| Gly-Pro p-nitroanilide hydrochloride >99% (TLC) |
Dipeptidyl peptidase IV |
G0513 |
| Gly-Pro-7-amido-4-methylcoumarin hydrobromide |
Dipeptidyl peptidase IV. |
G2761 |
| Gly-Pro p-nitroanilide p-toluenesulfonate salt ~99% (TLC) |
Dipeptidyl peptidase IV. |
G2901 |
| 2,3-Diphospho-D-glyceric acid penta(cyclohexylammonium) salt BioChemika ³97.0 % (HPCE) |
Diphosphoglycerate-independent phosphoglycerate mutase |
43380 |
| Tyramine BioChemika for fluorescence ³99.0% (TLC) |
Dopamine beta-hydroxylase |
93810 |
| N-(Methoxysuccinyl)-Ala-Ala-Pro-Val 4-nitroanilide BioChemika ³97.0 % (HPLC) |
Elastase |
65352 |
| N-Succinyl-Ala-Ala-Ala-p-nitroanilide |
Elastase |
S4760 |
| N-Succinyl-Ala-Ala-Ala-p-nitroanilide BioChemika ³99.0 % (enzymatic) |
Elastase |
85975 |
| Boc-D-Phe-Ala-Nle-p-nitroanilide |
Elastase, Hunan Granulocyte |
B8397 |
| Boc-Ala-Pro-Nva p-chlorothiobenzyl ester |
Elastase, leukocyte |
B2017 |
| N-Methoxysuccinyl-Ala-Ala-Pro-Val-7-amido-4-methylcoumarin |
Elastase, leukocyte |
M9771 |
| N-(Methoxysuccinyl)-Ala-Ala-Pro-Val 4-nitroanilide |
Elastase, leukocyte |
M4765 |
| N-Methoxysuccinyl-Ala-Ala-Pro-Val-7-amido-4-trifluoromethylcoumarin solid |
Elastase, leukocyte |
M4563 |
| N-Methoxysuccinyl-Ala-Ala-Pro-Val-7-amido-4-methylcoumarin |
Elastase, pancreatic |
M9771 |
| 7-Methoxycoumarin-4-acetyl [Ala7-(2,4-Dinitrophenyl)Lys9]-Bradykinin trifluoroacetate salt >94 % (HPLC) |
Endothelin-converting enzyme-1 |
B5181 |
| N-Dansyl-D-Ala-Gly-p-nitro-Phe-Gly ~95% (HPLC) |
Enkephalinase |
D2155 |
| Glutaryl-Ala-Ala-Phe-4-methoxy-b-naphthylamide |
Enkephalinase |
G3769 |
| Gly-Asp-Asp-Asp-Asp-Lys-b-naphthylamide ³95% (HPLC) |
Enterokinase |
G5261 |
| (2S,3S)-trans-3-Phenyl-2-oxiranylmethyl 4-nitrophenyl carbonate BioChemika ³98.0 % (NMR) |
Epoxy-Hydrolase |
04088 |
| 2',7'-Dichlorofluorescin diacetate BioChemika for fluorescence ³90% (HPLC) |
Esterase |
35847 |
| 8-Acetoxy-N,N,N',N',N'',N''-hexamethylpyrene-1,3,6-trisulfonamide BioChemika for fluorescence ³98.0% (TLC) |
Esterase |
52612 |
| 8-Acetoxypyrene-1,3,6-trisulfonic acid trisodium salt BioChemika for fluorescence ³98.0% (TLC) |
Esterase |
00879 |
| N-Acetyl-L-tyrosine ethyl ester monohydrate BioChemika ³99.0 % (sum of enantiomers, HPLC) |
Esterase |
01530 |
| 3-(2-Benzoxazolyl)umbelliferyl acetate BioChemika for fluorescence ³98.0% (HPLC) |
Esterase |
12832 |
| 5-Bromo-4-chloro-3-indolyl acetate |
Esterase |
B4377 |
| 5-Bromo-4-chloroindoxyl 1,3-diacetate BioChemika ³99.0 % (HPLC) |
Esterase |
16675 |
| 5-Bromoindoxyl acetate BioChemika ³98.0 % (AT) |
Esterase |
17400 |
| 8-Butyryloxypyrene-1,3,6-trisulfonic acid trisodium salt BioChemika for fluorescence ³95.0% (TLC) |
Esterase |
20800 |
| Paraoxon-ethyl ~90% oil |
Esterase |
D9286 |
| Fluorescein dibutyrate BioChemika for fluorescence ³98.0% (HPCE) |
Esterase |
46942 |
| Fluorescein dilaurate BioChemika for fluorescence ³97.0% (HPCE) |
Esterase |
46943 |
| Indoxyl acetate |
Esterase |
I3500 |
| 4-Methylumbelliferyl acetate BioChemika for fluorescence ³97.0% (TLC) |
Esterase |
46002 |
| 4-Methylumbelliferyl butyrate BioChemika for fluorescence ³95% (HPCE) |
Esterase |
19362 |
| Na-Benzoyl-L-arginine ethyl ester hydrochloride BioChemika ³99.0% (AT) |
Esterase |
12880 |
| Naphthol AS acetate BioChemika for histology ³99.0% (HPLC) |
Esterase |
70465 |
| Naphthol AS-D acetate BioChemika for histology ³95.0% (HPLC) |
Esterase |
70472 |
| Naphthol AS-D acetate BioChemika ³99.0 % (HPLC) |
Esterase |
70488 |
| Naphthol AS chloroacetate BioChemika for histology ³98.0% (HPLC) |
Esterase |
70470 |
| Naphthol AS-D chloroacetate BioChemika for histology ³95.0% (HPLC) |
Esterase |
70468 |
| Naphthol AS-D chloroacetate |
Esterase |
N0758 |
| Naphthol AS-E acetate BioChemika for histology ³96.0% (HPLC) |
Esterase |
70484 |
| Naphthol AS-MX acetate BioChemika for histology ~90% (HPLC) |
Esterase |
70475 |
| 1-Naphthyl acetate BioChemika ³99.0 % (HPLC) |
Esterase |
46010 |
| Primisulfuron-methyl PESTANAL®, analytical standard |
Esterase |
46010 |
| 2-Naphthyl acetate BioChemika substrate for the det. of esterase ³99.0 % (GC) |
Esterase |
46020 |
| 1-Naphthyl acetate |
Esterase |
N8505 |
| 1-Naphthyl butyrate |
Esterase |
N8000 |
| 1-Naphthyl butyrate BioChemika ³99.0 % (GC) |
Esterase |
19363 |
| 4-Nitrophenyl butyrate |
Esterase |
|
| 4-Nitrophenyl decanoate BioChemika ³99.0 % (TLC) |
Esterase |
21497 |
| 4-Nitrophenyl hexanoate BioChemika ³98.0 % (TLC) |
Esterase |
21610 |
| 4-Nitrophenyl octanoate BioChemika ³95.0 % (GC) |
Esterase |
21742 |
| 4-Nitrophenyl dodecanoate BioChemika ³98.0 % (GC) |
Esterase |
61716 |
| 4-Nitrophenyl myristate BioChemika ³95.0 % (HPLC) |
Esterase |
70124 |
| 4-Nitrophenyl palmitate BioChemika ³98.0 % (GC) |
Esterase |
76166 |
| 4-Nitrophenyl stearate BioChemika ³97.0 % (HPLC) |
Esterase |
85774 |
| 8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt BioChemika for fluorescence ³90% (HPCE) |
Esterase |
74875 |
| 8-Oleoyloxypyrene-1,3,6-trisulfonic acid trisodium salt BioChemika for fluorescence |
Esterase |
75344 |
| L-Phenylalanine ethyl ester hydrochloride puriss. ³99.0% (AT) |
Esterase |
78050 |
| Resorufin acetate BioChemika for fluorescence ³98.0% (TLC) |
Esterase |
83636 |
| 4-Methylumbelliferyl acetate |
Esterase probably |
M0883 |
| Thymidine 3'-phosphate 4-nitrophenyl ester sodium salt BioChemika ³80.0 % (HPLC) |
Exonuclease |
89298 |
| N-Benzoyl-Ile-Glu-Gly-Arg p-nitroanilide hydrochloride |
Factor X Activated (Xa) |
B2291 |
| N-Benzoyl-Ile-Glu-Gly-Arg p-nitroanilide acetate salt |
Factor X Activated (Xa) |
B7020 |
| Boc-Ile-Glu-Gly-Arg-7-amido-4-methylcoumarin hydrochloride |
Factor X Activated (Xa) |
B9760 |
| Factor Xa chromogenic substrate solid |
Factor X Activated (Xa) |
F3301 |
| 4-Methylumbelliferyl 4-guanidinobenzoate hydrochloride hydrate crystalline |
Factor X Activated (Xa) |
M4149 |
| 4-Methylumbelliferyl 4-guanidinobenzoate hydrochloride hydrate BioChemika for fluorescence |
Factor X Activated (Xa) |
51010 |
| 4-Nitrophenyl 4-guanidinobenzoate hydrochloride BioChemika ³95.0 % (AT) |
Factor X Activated (Xa) |
51015 |
| Boc-g-benzyl-Glu-Ala-Arg-7-amido-4-methylcoumarin hydrochloride ~95% |
Factor Xia |
B4903 |
| Boc-Phe-Ser-Arg-7-amido-4-methylcoumarin |
Factor XIA |
B6388 |
| Pefachrome® Fxa BioChemika ³95.0 % (HPLC) |
Faktor Xa |
77739 |
| 4-Methylumbelliferyl a-L-fucopyranoside ~98% (TLC) |
Fucosidase, alpha-L- |
M8527 |
| 4-Methylumbelliferyl a-L-fucopyranoside BioChemika for fluorescence ³98.0% (HPLC) |
Fucosidase, alpha-L- |
69588 |
| 4-Nitrophenyl a-L-fucopyranoside ³98% (TLC) |
Fucosidase, alpha-L- |
N3628 |
| 4-Nitrophenyl a-L-fucopyranoside BioChemika ³98.0 % (TLC) |
Fucosidase, alpha-L- |
73651 |
| 4-Nitrophenyl b-D-fucopyranoside BioChemika ³99.0 % (HPLC) |
Fucosidase, alpha-L- |
73652 |
| N-Acetyl-Arg-Glu-Lys-Arg-7-amido-4-trifluoromethylcoumarin >97 % (HPLC) |
Furin |
C6483 |
| 3-Nitrotetrazolium blue chloride BioChemika ³75 % (AT) |
Furoyl-CoA dehydrogenase, 2- |
09972 |
| Nitrotetrazolium blue chloride BioChemika ³88 % (AT) |
Furoyl-CoA dehydrogenase, 2- |
74030 |
| Nitrotetrazolium blue chloride BioChemika ~10 mg/tablet nitrotetrazolium blue chloride |
Furoyl-CoA dehydrogenase, 2- |
74031 |
| Nitrotetrazolium blue chloride BioChemika ³98.0 % (calc. based on dry substance, AT) |
Furoyl-CoA dehydrogenase, 2- |
74032 |
| Tetrazolium Blue Chloride BioChemika indicator for germination, for microbiology ³90% (AT) |
Furoyl-CoA dehydrogenase, 2- |
88190 |
| 4-Methylumbelliferyl N-acetyl-b-D-galactosaminide ³98% (TLC) |
Galactosaminidase, beta |
M9659 |
| 5-Bromo-4-chloro-3-indolyl N-acetyl-b-D-galactosaminide ~95% |
Galactosaminidase, N-acetyl-beta-D |
B3166 |
| 5-Bromo-4-chloro-3-indolyl N-acetyl-b-D-galactosaminide BioChemika ³98.0 % (TLC) |
Galactosaminidase, N-acetyl-beta-D |
16658 |
| 4-Nitrophenyl-N-acetyl-a-D-galactosaminide BioChemika ³99.0 % (TLC) |
Galactosaminidase, N-acetyl-beta-D |
73603 |
| 4-Nitrophenyl N-acetyl-b-D-galactosaminide BioChemika ³99.0 % (TLC) |
Galactosaminidase, N-acetyl-beta-D |
73604 |
| 4-Nitrophenyl a-D-galactopyranoside |
Galactosidase, alpha |
N0877 |
| 4-Nitrophenyl a-D-galactopyranoside BioChemika ³99.0 % (HPLC) |
Galactosidase, alpha |
73653 |
| 4-Methylumbelliferyl a-D-galactopyranoside ³98% (TLC) |
Galactosidase, alpha- |
M7633 |
| 4-Methylumbelliferyl a-D-glucopyranoside |
Galactosidase, alpha- |
M9766 |
| 4-Methylumbelliferyl a-D-galactopyranoside BioChemika for fluorescence ³96.0% (TLC) |
Galactosidase, alpha- |
69586 |
| 4-Trifluoromethylumbelliferyl b-D-galactopyranoside BioChemika for fluorescence ³95% (HPCE) |
Galactosidase, beta |
91879 |
| 5-Bromo-4-chloro-3-indolyl b-D-galactopyranoside BioChemika for molecular biology ³99.0% (TLC) |
Galactosidase, beta |
16664 |
| 5-Bromo-4-chloro-3-indolyl b-D-galactopyranoside BioChemika ³98.0 % (HPLC) |
Galactosidase, beta |
16665 |
| 5-Bromo-6-chloro-3-indolyl-b-D-galactopyranoside BioChemika ³98.0 % (HPLC) |
Galactosidase, beta |
16669 |
| 5-Bromo-3-indolyl b-D-galactopyranoside BioChemika ³98.0 % (HPLC) |
Galactosidase, beta |
17395 |
| 6-Bromo-2-naphthyl b-D-galactopyranoside BioChemika ³98.0 % (HPLC) |
Galactosidase, beta |
17664 |
| 6-Chloro-3-indolyl-b-D-galactopyranoside BioChemika ³98.0 % (HPLC) |
Galactosidase, beta |
24904 |
| 6,8-Difluoro-4-methylumbelliferyl-b-D-galactopyranoside BioChemika for fluorescence ³98.0% (TLC) |
Galactosidase, beta |
36931 |
| Fluorescein di(b-D-galactopyranoside) BioChemika for fluorescence ³98.0% (TLC) |
Galactosidase, beta |
46941 |
| Fluorescein di-(b-D-N-acetylgalactosamine) |
Galactosidase, beta |
F0802 |
| 5-Iodo-3-indolyl-b-D-galactopyranoside BioChemika ³98.0 % (HPLC) |
Galactosidase, beta |
57897 |
| 1-Methyl-3-indolyl-b-D-galactopyranoside BioChemika ³98.0 % (HPLC) |
Galactosidase, beta |
67610 |
| 4-Methylumbelliferyl-b-D-galactopyranoside BioChemika for fluorescence ³97.0% (HPLC) |
Galactosidase, beta |
69590 |
| 4-Methylumbelliferyl b-D-lactopyranoside BioChemika for fluorescence ³95% (HPCE) |
Galactosidase, beta |
69601 |
| 2-Naphthyl b-D-galactopyranoside BioChemika ³99.0 % (TLC) |
Galactosidase, beta |
70965 |
| 4-Nitrophenyl b-D-galactopyranoside ³98% (enzymatic) |
Galactosidase, beta |
N1252 |
| 2-Nitrophenyl b-D-galactopyranoside BioChemika ³99.0 % (HPLC) |
Galactosidase, beta |
73660 |
| 4-Nitrophenyl b-D-galactopyranoside BioChemika ³99.0 % (HPLC) |
Galactosidase, beta |
73670 |
| Phenyl-b-D-galactopyranoside BioChemika ³99.0 % (HPLC) |
Galactosidase, beta |
78552 |
| Resorufin b-D-galactopyranoside BioChemika for fluorescence ³95% (HPLC) |
Galactosidase, beta |
83638 |
| 5-Bromo-4-chloro-3-indolyl b-D-galactopyranoside ³98% powder |
Galactosidase, beta- |
B4252 |
| 5-Bromo-4-chloro-3-indolyl b-D-galactopyranoside tablet |
Galactosidase, beta- |
B6024 |
| 5-Bromo-3-indolyl b-D-galactopyranoside ³98% |
Galactosidase, beta- |
B4387 |
| 2-Chloro-4-nitrophenyl-b-D-lactoside BioChemika chromophoric enzyme substrate ³90 % (HPCE) |
Galactosidase, beta- |
25589 |
| Fluorescein di(b-D-galactopyranoside) |
Galactosidase, beta- |
F2756 |
| Fluorescein mono-b-D-galactopyranoside ~90% |
Galactosidase, beta- |
F6542 |
| 1-Methyl-3-indolyl-b-D-galactopyranoside ³98.0 % (HPLC) solid |
Galactosidase, beta- |
M1688 |
| 4-Methylumbelliferyl-b-D-galactopyranoside ³99% (TLC) |
Galactosidase, beta- |
M1633 |
| Pyridoxine galactoside |
Galactosidase, beta- |
P6239 |
| RH 421 ³95 % solid |
Galactosidase, beta- |
R8026 |
| [2-Aminobenzoyl-Gly1]-Galanin fragment (1-10)-Lys(retro-m-Nitro-Tyr) amide, human trifluoroacetate salt >95 % (HPLC) solid |
Galanin degrading zinc-metallopeptidase |
G8916 |
| Rac-1 protein human ³85 % recombinant, expressed in Escherichia coli buffered aqueous glycerol solution |
Geranylgeranyltransferase |
R3012 |
| Ras, modified human ~95% (SDS-PAGE) recombinant, expressed in Escherichia coli buffered aqueous glycerol solution |
Geranylgeranyltransferase |
R0145 |
| Laminarin from Laminaria digitata BioChemika |
Glucanase, beta- |
61430 |
| Cellopentaose BioChemika ³85% (HPLC) |
Glucanase, endo-1,4- |
22163 |
| Ostazin Brilliant Red H-3B hydroxyethyl cellulose |
Glucanase, endo-1,4- |
O6879 |
| Hydroxyethyl-cellulose dyed with ostazin brilliant red H-3B BioChemika 10 % as dye |
Glucanases, endo-1,4-beta- |
54295 |
| 5-Bromo-4-chloro-3-indolyl-N-acetyl-b-D-glucosaminide BioChemika ³98.0 % (TLC) |
Glucosaminidase, N-acetyl-beta- |
16659 |
| 4-Methylumbelliferyl N-acetyl-b-D-glucosaminide dihydrate BioChemika for fluorescence ³99.0% (TLC) |
Glucosaminidase, N-acetyl-beta- |
69585 |
| 4-Nitrophenyl N-acetyl-b-D-glucosaminide BioChemika ³99.0 % (TLC) |
Glucosaminidase, N-acetyl-beta- |
73606 |
| 3-Nitrotetrazolium blue chloride BioChemika ³75 % (AT) |
Glucose Oxidase |
09972 |
| Nitrotetrazolium blue chloride BioChemika ³88 % (AT) |
Glucose Oxidase |
74030 |
| Nitrotetrazolium blue chloride BioChemika ~10 mg/tablet nitrotetrazolium blue chloride |
Glucose Oxidase |
74031 |
| Nitrotetrazolium blue chloride BioChemika ³98.0 % (calc. based on dry substance, AT) |
Glucose Oxidase |
74032 |
| Tetranitroblue tetrazolium chloride BioChemika ³85 % (AT) |
Glucose Oxidase |
87961 |
| Tetrazolium Blue Chloride BioChemika indicator for germination, for microbiology ³90% (AT) |
Glucose Oxidase |
88190 |
| 3-Nitrotetrazolium blue chloride BioChemika ³75 % (AT) |
Glucose-6-phosphate dehydrogenase |
09972 |
| Nitrotetrazolium blue chloride BioChemika ³88 % (AT) |
Glucose-6-phosphate dehydrogenase |
74030 |
| Nitrotetrazolium blue chloride BioChemika ~10 mg/tablet nitrotetrazolium blue chloride |
Glucose-6-phosphate dehydrogenase |
74031 |
| Nitrotetrazolium blue chloride BioChemika ³98.0 % (calc. based on dry substance, AT) |
Glucose-6-phosphate dehydrogenase |
74032 |
| Tetrazolium Blue Chloride BioChemika indicator for germination, for microbiology ³90% (AT) |
Glucose-6-phosphate dehydrogenase |
88190 |
| 4-Methylumbelliferyl a-D-glucopyranoside BioChemika for fluorescence ³99.0% (TLC) |
Glucosidase, alpha |
69591 |
| 4-Nitrophenyl a-D-glucopyranoside ³99% |
Glucosidase, alpha |
N1377 |
| 4-Nitrophenyl a-D-glucopyranoside BioChemika ³98.0 % (HPLC) |
Glucosidase, alpha |
73673 |
| 4-Nitrophenyl a-D-maltohexaoside BioChemika ³98.0 % (HPLC) |
Glucosidase, alpha |
73681 |
| 4-Nitrophenyl a-D-maltopentaoside BioChemika ³98.0 % (HPLC) |
Glucosidase, alpha |
73734 |
| 4-Nitrophenyl-6'''''-deoxy-6'''''-(2-pyridylamino)-a-D-penta-(1®4)- glucopyranoside BioChemika ³95 % (HPLC) |
Glucosidase, alpha |
73618 |
| 4-(Trifluoromethyl)umbelliferyl-b-D-glucopyranoside BioChemika for fluorescence ³95% (HPCE) |
Glucosidase, beta- |
91880 |
| 3-Acetylumbelliferyl b-D-glucopyranoside ³95% |
Glucosidase, beta- |
A3582 |
| 5-Bromo-4-chloro-3-indolyl b-D-glucopyranoside ~98% |
Glucosidase, beta- |
B4527 |
| 5-Bromo-4-chloro-3-indolyl b-D-glucopyranoside BioChemika ³99.0 % (TLC) |
Glucosidase, beta- |
16667 |
| 2-Chloro-4-nitrophenyl-b-D-glucopyranoside BioChemika chromophoric enzyme substrate ³98.0 % (TLC) |
Glucosidase, beta- |
25588 |
| 6,8-Difluoro-4-methylumbelliferyl-b-D-glucopyranoside BioChemika for fluorescence ³98.0% (TLC) |
Glucosidase, beta- |
36937 |
| Fluorescein di(b-D-glucuronide) |
Glucosidase, beta- |
F4551 |
| 4-Methylumbelliferyl b-D-glucopyranoside |
Glucosidase, beta- |
M3633 |
| 4-Methylumbelliferyl b-D-glucopyranoside BioChemika for fluorescence ³99.0% (HPLC) |
Glucosidase, beta- |
69600 |
| 4-Nitrophenyl b-D-glucopyranoside |
Glucosidase, beta- |
N7006 |
| 2-Nitrophenyl b-D-glucopyranoside BioChemika ³99.0 % (HPLC) |
Glucosidase, beta- |
73674 |
| 4-Nitrophenyl b-D-glucopyranoside BioChemika ³99.0 % (HPLC) |
Glucosidase, beta- |
73676 |
| Phenyl b-D-glucopyranoside BioChemika ³98.0 % (HPLC) |
Glucosidase, beta- |
78554 |
| Resorufin b-D-glucopyranoside BioChemika for fluorescence ³95% (HPLC) |
Glucosidase, beta- |
83639 |
| D-(-)-Salicin ³99% |
Glucosidase, beta- |
S0625 |
| 5-Bromo-4-chloro-3-indolyl b-D-cellobioside |
Glucosidases, beta- (selected) |
B6795 |
| 5-Bromo-4-chloro-3-indolyl--D-glucuronide Sodium salt |
Glucuronidase, beta- |
|
| 5-Bromo-4-chloro-3-indolyl b-D-glucuronide cyclohexylammonium salt BioChemika for molecular biology ³99.0% (TLC) |
Glucuronidase, beta- |
16666 |
| 5-Bromo-6-chloro-3-indolyl b-D-glucuronide cyclohexylammonium salt BioChemika ³98.0 % (HPLC) |
Glucuronidase, beta- |
16671 |
| 6-Chloro-3-indolyl-b-D-glucuronide cyclohexylamine salt BioChemika ³97.0 % (HPLC) |
Glucuronidase, beta- |
24907 |
| 6,8-Difluoro-4-methylumbelliferyl-b-D-glucuronide lithium salt BioChemika for fluorescence ³98.0% (TLC) |
Glucuronidase, beta- |
36939 |
| 4-Methylumbelliferyl-b-D-glucuronide trihydrate BioChemika for fluorescence ³99.0% (HPLC) |
Glucuronidase, beta- |
69602 |
| Naphthol AS-BI b-D-glucuronide |
Glucuronidase, beta- |
N1875 |
| Naphthol AS-BI b-D-glucuronide BioChemika for histology ³99.0% (TLC) |
Glucuronidase, beta- |
70493 |
| 4-Nitrophenyl b-D-glucuronide BioChemika ³99.0 % (TLC) |
Glucuronidase, beta- |
73677 |
| Phenolphthalein b-D-glucuronide sodium salt rabbit urine |
Glucuronidase, beta- |
P0376 |
| Phenolphthalein glucuronic acid sodium salt dihydrate rabbit urine BioChemika ³98.0 % (TLC) |
Glucuronidase, beta- |
77637 |
| Phenolphthalein b-D-glucuronide from rabbit urine BioChemika ³98.0 % (TLC) |
Glucuronidase, beta- |
77636 |
| Phenyl-b-D-glucuronide BioChemika shipped in dry ice ³99.0 % (HPLC) |
Glucuronidase, beta- |
78555 |
| Resorufin b-D-glucuronide sodium salt ³90% |
Glucuronidase, beta- |
R2886 |
| 5-Bromo-4-chloro-3-indolyl b-D-glucuronide cyclohexylammonium salt ³98% |
Glucuronidase, beta- (selection) |
B0522 |
| 5-Bromo-4-chloro-3-indolyl b-D-glucuronide cyclohexylammonium salt ³98% |
Glucuronidase, beta- (selection) |
B3783 |
| 5-Bromo-4-chloro-3-indolyl b-D-glucuronide cyclohexylammonium salt tablet |
Glucuronidase, beta- bacterial |
B8049 |
| 5-Bromo-4-chloro-3-indolyl b-D-glucuronide sodium salt tablet |
Glucuronidase, beta- bacterial |
B8174 |
| 5-Bromo-6-chloro-3-indolyl b-D-glucuronide cyclohexylammonium salt ³98 % |
Glucuronidase, beta- bacterial |
B4532 |
| 6-Bromo-2-naphthyl b-D-glucuronide |
Glucuronidase, beta- bacterial |
B6519 |
| 8-Hydroxyquinoline glucuronide 95-100% powder |
Glucuronidase, beta- bacterial |
H1254 |
| 4-Methylumbelliferyl b-D-glucuronide hydrate SigmaUltra |
Glucuronidase, beta- bacterial |
M5664 |
| a-Ketoglutaric acid ³98.5% (NaOH, titration) |
Glutamic dehydrogenase, L- |
K1750 |
| a-Ketoglutaric acid potassium salt |
Glutamic dehydrogenase, L- |
K2000 |
| a-Ketoglutaric acid sodium salt |
Glutamic dehydrogenase, L- |
K1875 |
| a-Ketoglutaric acid sodium salt SigmaUltra |
Glutamic dehydrogenase, L- |
K2010 |
| Z-Leu-Leu-Glu b-naphthylamide ³98% powder |
Glutamylpeptidyl-peptide hydrolase |
C0788 |
| L-Glutamic acid g-(3-carboxy-4-nitroanilide) ammonium salt BioChemika ³99.0 % (TLC) |
Glutamyltransferase, gamma- |
49525 |
| L-Glutamic acid g-(4-nitroanilide) BioChemika ³99.0 % (NT) |
Glutamyltransferase, gamma- |
49623 |
| Cumene hydroperoxide ~80 % |
Glutathione Peroxidase |
C0524 |
| 1-Chloro-2,4-dinitrobenzene ³98 % |
Glutathione S-transferase |
C6396 |
| 4-Chloro-7-nitrobenzofurazan BioChemika for fluorescence ³99.0% (HPLC) puriss. |
Glutathione S-transferase |
25455 |
| sn-Glycerol 3-phosphate bis(cyclohexylammonium) salt BioChemika ³95 % (GC) |
Glycerophosphate dehydrogenase, alpha- |
49990 |
| 4-Nitrophenyl b-D-fucopyranoside ³98% (TLC) |
Glycosidase, beta |
N3378 |
| Z-Gly-Pro-Arg 7-amido-4-methylcoumarin hydrobromide ³85 % (TLC) powder |
Granzyme A |
C0855 |
| Z-Gly-Pro-Arg 7-amido-4-trifluoromethylcoumarin ³97 % (HPLC) powder |
Granzyme A |
C0980 |
| Z-Gly-Pro-Arg 4-methoxy-2-naphthylamide ³97 % (HPLC) powder |
Granzyme A |
C1105 |
| N-Acetyl-Ile-Glu-Pro-Asp-7-amido-4-methylcoumarin ³97 % (HPLC) powder |
Granzyme B |
A6220 |
| N-Acetyl-Ile-Glu-Pro-Asp-p-nitroaniline ³97 % (HPLC) powder |
Granzyme B |
A6470 |
| N-Acetyl-Ile-Glu-Thr-Asp-p-nitroanilide ³95 % |
Granzyme B |
A9968 |
| N-Acetyl-Ile-Glu-Thr-Asp-7-Amido-4-methylcoumarin ³97 % (HPLC) powder |
Granzyme B |
A4188 |
| Boc-Ala-Ala-Asp thiobenzyl ester ³97 % (HPLC) powder |
Granzyme B |
B5305 |
| Z-Ile-Glu-Thr-Asp 7-amido-4-trifluoromethylcoumarin ³95 % (HPLC) |
Granzyme B |
C5599 |
| N-Acetyl-Ile-Glu-Thr-Asp-p-nitroanilide ³95 % |
Granzyme B |
A9968 |
| Z-Ile-Glu-Thr-Asp 7-amido-4-trifluoromethylcoumarin ³95 % (HPLC) |
Granzyme B |
C5599 |
| Guanosine 5'-triphosphate P3-[1-(2-nitrophenyl)ethyl ester] disodium salt BioChemika ³95 % (HPLC) |
GTPase |
51125 |
| Histamine dihydrochloride BioChemika ³99.0 % (AT) |
Histamine-N-methyltransferase |
53300 |
| Iodonitrotetrazolium chloride BioChemika for histol. analysis (of oxidases) ³97.0% (AT) |
Histidinol dehydrogenase |
58030 |
| Histone Deacetylase Substrate (Fluorogenic) >98 % (HPLC) crystalline solid |
Histone Deacetylase |
H9660 |
| Anthranilyl-His-Lys-Ala-Arg-Val-Leu-p-nitro-Phe-Glu-Ala-Nle-Ser amide |
HIV Protease |
A0811 |
| Anthranilyl-HIV Protease Substrate >94 % (HPLC) |
HIV Protease |
H5910 |
| Anthranilyl-HIV Protease Substrate V >95 % (HPLC) |
HIV Protease |
H6035 |
| Anthranilyl-HIV Protease Substrate VI >95 % (HPLC) |
HIV Protease |
H6160 |
| HIV Protease Substrate 1 |
HIV Protease |
H6660 |
| HIV Protease Substrate III-B, Native Sequence >98 % (HPLC) |
HIV Protease |
H5285 |
| HIV Protease Substrate V >98 % (HPLC) |
HIV Protease |
H5410 |
| HIV Protease Substrate VI >97 % (HPLC) |
HIV Protease |
H5535 |
| HIV Protease Substrate VII >97 % (HPLC) |
HIV Protease |
H5660 |
| HIV Protease Substrate VIII >96 % (HPLC) |
HIV Protease |
H5785 |
| Boc-Val-Leu-Gly-Arg p-nitroanilide hydrobromide |
Horseshoe crab clotting enzyme |
B8391 |
| 3-(N-Ethyl-3-methylanilino)-2-hydroxypropanesulfonic acid sodium salt purum p.a. for enzymic, spectrophotometric determination of H2O2 ³98.0 % (dried material, NT) |
Hydrogenperoxide |
04340 |
| 4-Methylumbelliferyl a-L-iduronide cyclohexylammonium salt ³99% (TLC) |
Iduronidase |
M9531 |
| Interleukin-1b precursor fragment 110-123 ³97 % (HPLC) |
Interleukin-1beta converting enzyme |
I2775 |
| 3-Nitrotetrazolium blue chloride BioChemika ³75 % (AT) |
Isoquinoline 1-oxidoreductase |
09972 |
| Nitrotetrazolium blue chloride BioChemika ³88 % (AT) |
Isoquinoline 1-oxidoreductase |
74030 |
| Nitrotetrazolium blue chloride BioChemika ~10 mg/tablet nitrotetrazolium blue chloride |
Isoquinoline 1-oxidoreductase |
74031 |
| Nitrotetrazolium blue chloride BioChemika ³98.0 % (calc. based on dry substance, AT) |
Isoquinoline 1-oxidoreductase |
74032 |
| Tetrazolium Blue Chloride BioChemika indicator for germination, for microbiology ³90% (AT) |
Isoquinoline 1-oxidoreductase |
88190 |
| N-Benzoyl-Pro-Phe-Arg-p-nitroanilide hydrochloride |
Kallikrein |
B2133 |
| Z-Phe-Arg 7-amido-4-methylcoumarin hydrochloride |
Kallikrein |
C9521 |
| Kininogen, low molecular weight from human plasma >95% (SDS-PAGE) lyophilized powder |
Kallikrein |
K3628 |
| Na-Benzoyl-L-arginine methyl ester bicarbonate salt |
Kallikrein |
B4256 |
| 4-Nitrophenyl 4-guanidinobenzoate hydrochloride BioChemika ³95.0 % (AT) |
Kallikrein |
51015 |
| Na-p-Tosyl-L-arginine methyl ester hydrochloride BioChemika for the determination of trypsin ³99.0 % (AT) |
Kallikrein |
90170 |
| D-Val-Leu-Arg 7-amido-4-methylcoumarin solid |
Kallikrein |
V2763 |
| D-Val-Leu-Arg 7-amido-4-trifluoromethylcoumarin ³90 % (HPLC) solid |
Kallikrein |
V2888 |
| a-Ketoglutaric acid ³98.5% (NaOH, titration) |
Ketoglutarate Dehydrogenase, alpha |
K1750 |
| a-Ketoglutaric acid potassium salt |
Ketoglutarate Dehydrogenase, alpha |
K2000 |
| a-Ketoglutaric acid sodium salt |
Ketoglutarate Dehydrogenase, alpha |
K1875 |
| a-Ketoglutaric acid sodium salt SigmaUltra |
Ketoglutarate Dehydrogenase, alpha |
K2010 |
| Iodonitrotetrazolium chloride BioChemika for histol. analysis (of oxidases) ³97.0% (AT) |
Ketolisomerase |
58030 |
| Iodonitrotetrazolium chloride BioChemika for histol. analysis (of oxidases) ³97.0% (AT) |
Lactate dehydrogenase |
58030 |
| 4-[3-(4-Iodophenyl)-2-(2,4-dinitrophenyl)-2H-5-tetrazolio]-1,3-benzenedisulfonate sodium salt BioChemika ³90 % (HPLC) |
Lactic Dehydrogenase |
90482 |
| 3-Nitrotetrazolium blue chloride BioChemika ³75 % (AT) |
Lactic Dehydrogenase |
09972 |
| Nitrotetrazolium blue chloride BioChemika ³88 % (AT) |
Lactic Dehydrogenase |
74030 |
| Nitrotetrazolium blue chloride BioChemika ~10 mg/tablet nitrotetrazolium blue chloride |
Lactic Dehydrogenase |
74031 |
| Nitrotetrazolium blue chloride BioChemika ³98.0 % (calc. based on dry substance, AT) |
Lactic Dehydrogenase |
74032 |
| Sodium 2-oxobutyrate BioChemika ³99.0 % (NT) |
Lactic Dehydrogenase |
75825 |
| a-Ketoglutaric acid sodium salt BioChemika ³98.0 % (NT) |
Lactic Dehydrogenase |
75893 |
| Pyruvic acid ³90 % (enzymatic) liquid |
Lactic Dehydrogenase |
P1656 |
| Tetranitroblue tetrazolium chloride BioChemika ³85 % (AT) |
Lactic Dehydrogenase |
87961 |
| Tetrazolium Blue Chloride BioChemika indicator for germination, for microbiology ³90% (AT) |
Lactic Dehydrogenase |
88190 |
| Iodonitrotetrazolium chloride BioChemika for histol. analysis (of oxidases) ³97.0% (AT) |
Lactyl-CoA dehydrogenase |
58030 |
| Laminarin azure from Laminaria digitata |
Laminarinase |
L0145 |
| L-Leucine-p-nitroanilide |
Leucine aminopeptidase |
L9125 |
| L-Leucine 7-amido-4-methyl coumarin hydrochloride BioChemika for fluorescence |
Leucine aminopeptidase |
61888 |
| L-Leucine b-naphthylamide hydrochloride BioChemika substrate for the det. of leucine aminopeptidase acc. Seligmann ³97.0 % (AT) |
Leucine aminopeptidase |
61900 |
| N-Succinyl-Ala-Ala-Pro-Phe-7-amido-4-methylcoumarin |
Leucine Aminopeptidase |
S9761 |
| N-Succinyl-Phe-Gly-Leu-b-naphthylamide |
LH-RH-degrading endopeptidase |
S6768 |
| Boc-Ser(Bzl)-Gly-Arg-NH-Np.HCl BioChemika ³98.0 % (HPLC) |
Limulus Clotting Enzyme |
76556 |
| 1,2-Di-O-decanoyl-rac-glycero-3-glutaric acid-resorufin labeled |
Lipase |
D7414 |
| Fluorescein dibutyrate BioChemika for fluorescence ³98.0% (HPCE) |
Lipase |
46942 |
| Fluorescein dilaurate BioChemika for fluorescence ³97.0% (HPCE) |
Lipase |
46943 |
| Indoxyl acetate BioChemika ³97.0% (TLC) |
Lipase |
57420 |
| Lipase Substrate BioChemika for the titrimetric determination of enzyme activity |
Lipase |
62314 |
| 4-Methylumbelliferyl butyrate BioChemika for fluorescence ³95% (HPCE) |
Lipase |
19362 |
| 4-Methylumbelliferyl heptanoate BioChemika for fluorescence ³98.0% (TLC) |
Lipase |
75230 |
| 4-Methylumbelliferyl oleate BioChemika for fluorescence ³90% (HPCE) |
Lipase |
75164 |
| 4-Methylumbelliferyl palmitate BioChemika for fluorescence ³98.0% (TLC) |
Lipase |
76163 |
| 2-Naphthyl laurate BioChemika substrate for the det. of lipase ³99.0 % (GC) |
Lipase |
61710 |
| Resorufin butyrate BioChemika for fluorescence ³95% (TLC) |
Lipase |
83637 |
| 4-Trifluoromethylumbelliferyl oleate |
Lipase |
T5069 |
| Alizarin-3-methyliminodiacetic acid |
Lipid Peroxidation |
A3882 |
| Firefly tails (lanterns), desiccated |
Luciferase |
FFT |
| Fireflies, whole, desiccated |
Luciferase |
FFW |
| Firefly Lantern Extract powder |
Luciferase |
F3641 |
| Firefly Lantern Extract |
Luciferase |
F8021 |
| FLE50 FIREFLY LANTERN EXTRACT |
Luciferase |
FLE-50 |
| Firefly diluent lyophilized powder |
Luciferase |
F3766 |
| D-Luciferin BioChemika ³96.0 % (HPLC) |
Luciferase |
62643 |
| Luciferin 6'-ethyl ether sodium salt |
Luciferase |
L9007 |
| Luciferin 6'-methyl ether sodium salt |
Luciferase |
L1147 |
| D-Luciferin synthetic SigmaUltra ³99% (HPLC) |
Luciferase |
L6152 |
| D-Luciferin sodium salt lyophilized powder synthetic |
Luciferase |
L6882 |
| D-Luciferin sodium salt BioChemika ³98.0 % (HPLC) |
Luciferase |
62645 |
| D-Luciferin synthetic |
Luciferase |
L9504 |
| D-Luciferin potassium salt |
Luciferase |
L7538 |
| Peptidoglycan dyed with Remazol Brilliant Blue R Staphylococcus aureus BioChemika |
Lysostaphin |
77145 |
| Peptidoglycan from Micrococcus luteus BioChemika |
Lysostaphin |
53243 |
| 4-Methylumbelliferyl b-D-N,N',N''-triacetylchitotrioside |
Lysozyme |
M5639 |
| 4-Methylumbelliferyl b-D-N,N',N''-triacetylchitotrioside BioChemika for fluorescence 90% |
Lysozyme |
69615 |
| 4-Nitrophenyl b-D-N,N',N''-triacetylchitotriose |
Lysozyme |
N8638 |
| Peptidoglycan dyed with Remazol Brilliant Blue R Staphylococcus aureus BioChemika |
Lysozyme |
77145 |
| Peptidoglycan from Micrococcus luteus BioChemika |
Lysozyme |
53243 |
| Yeast BioChemika powder color slightly brown |
Lyticase |
51475 |
| Oxaloacetic acid BioChemika ³98.0 % (T) |
Malate dehydrogenase |
75660 |
| 4-Methylumbelliferyl a-D-mannopyranoside ~98% (TLC) |
Mannosidase, alpha- |
M3657 |
| 4-Methylumbelliferyl a-D-mannopyranoside BioChemika for fluorescence ³99.0% (TLC) |
Mannosidase, alpha- |
69604 |
| 4-Nitrophenyl a-D-mannopyranoside BioChemika ³99.0 % (TLC) |
Mannosidase, alpha- |
73726 |
| 4-Nitrophenyl a-D-mannopyranoside |
Mannosidase, alpha- |
N2127 |
| 4-Nitrophenyl b-D-mannopyranoside ³98% |
Mannosidase, Beta- |
N1268 |
| 4-Nitrophenyl b-D-mannopyranoside BioChemika ³99.0 % (TLC) |
Mannosidase, Beta- |
73727 |
| N-(2,4-Dinitrophenyl)-Pro-Leu-Gly-Met-Trp-Ser-Arg solid |
Matrix metalloproteinase |
D4315 |
| 7-Methoxycoumarin-4-acetyl-Pro-Leu-Gly-Leu-b-(2,4-dinitrophenylamino)Ala-Ala-Arg amide ³97% (HPLC) |
Matrix metalloproteinase |
M6412 |
| 7-Methoxycoumarin-4-acetyl-Pro-Leu-Ala-Nva-L-(2,4-dinitrophenyl)diaminopropio nyl-Ala-Arg-amide solid |
Matrix metalloproteinase-3 |
M3439 |
| N-(2,4-Dinitrophenyl)-Pro-Leu-Gly-Leu-Trp-Ala-D-Arg amide |
Matrix metalloproteinases MMP-1 and MMP-9 |
D2293 |
| N-(2,4-Dinitrophenyl)-Pro-Leu-Gly-Leu-Trp-Ala-D-Arg amide |
Matrix metalloproteinases MMP-1 and MMP-9 |
D2293 |
| DABCYL-[Nle1077]-Collagen Type III a 1 Chain fragment 1070-1077–EDANS, Human >96 % (HPLC) Type III a 1 chain |
Metalloproteinase |
C9732 |
| 7-Methoxycoumarin-4-acetyl-Arg-Pro-Leu-Ala-Leu-Trp-Arg-L-a,b-diaminopropionyl(2,4-dinitrophenyl) amide >97 % (HPLC) solid |
Metalloproteinase |
M0813 |
| N-Acetyl-S-geranylgeranyl-L-cysteine ³95% ethanol solution |
Methyltransferase, S-Adenosylmethionine-dependent |
A6331 |
| Kynuramine dihydrobromide crystalline |
Monoamine Oxidase |
K3250 |
| Tryptamine hydrochloride BioChemika ³98.0 % (AT) |
Monoamine oxidase |
93650 |
| Tyramine BioChemika for fluorescence ³99.0% (TLC) |
Monoamine oxidase |
93810 |
| 7-Ethoxycoumarin BioChemika for fluorescence ³99.0% (HPLC) |
Monooxygenase |
02644 |
| 1-Carbamoylmethyl-3-carbamoyl-1,4-dihydropyrimidine |
NAD(P)H quinone oxidoreductase 2 |
C1485 |
| CB 1954 powder |
NAD(P)H quinone oxidoreductase 2 |
C2235 |
| 1-(3-Sulfonatopropyl)-3-carbamoyl-1,4-dihydropyrimidine ³85 % (TLC) solid |
NAD(P)H quinone oxidoreductase 2 |
S0190 |
| 1-Naphthyl acetate BioChemika ³99.0 % (HPLC) |
Naphthyl Acetate Esterase |
46010 |
| Primisulfuron-methyl PESTANAL®, analytical standard |
Naphthyl Acetate Esterase |
46010 |
| 2-Naphthyl acetate BioChemika substrate for the det. of esterase ³99.0 % (GC) |
Naphthyl Acetate Esterase |
46020 |
| 5-Bromo-4-chloro-3-indolyl a-D-N-acetylneuraminic acid sodium salt ³90% |
Neuraminidase |
B4666 |
| 2'-(4-Methylumbelliferyl)-a-D-N-acetylneuraminic acid sodium salt hydrate ³95% (HPLC) |
Neuraminidase |
M8639 |
| 2'-(4-Methylumbelliferyl)-a-D-N-acetylneuraminic acid sodium salt hydrate BioChemika for fluorescence ³97.0% (HPLC) |
Neuraminidase |
69587 |
| 2-O-(p-Nitrophenyl)-a-D-N-acetylneuraminic acid ³95% |
Neuraminidase |
N1516 |
| L-Arginine reagent grade ³98% (TLC) powder |
Nitric Oxide Synthase |
A5006 |
| L-Arginine L-glutamate salt |
Nitric Oxide Synthase |
A5756 |
| L-Arginine monohydrochloride reagent grade ³98% (TLC) powder |
Nitric Oxide Synthase |
A5131 |
| NITRIC OXIDE SYNTHASE DETECTION SYSTEM, FLUOROMETRIC, FOR CELL-ASSOCIATED NOS |
NITRIC OXIDE SYNTHASE |
FCANOS-1 |
| NOC-5 ³95% |
Nitric Oxide Synthase/NO Donor |
A5456 |
| 3,3-Bis(aminoethyl)-1-hydroxy-2-oxo-1-triazene ³95% |
Nitric Oxide Synthase/NO Donor |
A5581 |
| Diethylamine NONOate diethylammonium salt |
Nitric Oxide Synthase/NO Donor |
D5431 |
| Diethylamine NONOate sodium salt crystalline |
Nitric Oxide Synthase/NO Donor |
D184 |
| Dipropylenetriamine NONOate |
Nitric Oxide Synthase/NO Donor |
D5057 |
| (±)-(E)-4-Ethyl-2-[(E)-hydroxyimino]-5-nitro-3-hexenamide ³98 % |
Nitric Oxide Synthase/NO Donor |
E2895 |
| (±)-(E)-4-Ethyl-2-[(Z)-hydroxyimino]-5-nitro-3-hexen-1-yl-nicotinamide ³97 % |
Nitric Oxide Synthase/NO Donor |
E3020 |
| (±)-(E)-2-[(E)-Hydroxyimino]-6-methoxy-4-methyl-5-nitro-3-hexenamide ³95 % (HPLC) |
Nitric Oxide Synthase/NO Donor |
H9528 |
| 1-Hydroxy-3-methyl-3-(methylaminopropyl)-2-oxo-1-triazene ³90% (HPLC) |
Nitric Oxide Synthase/NO Donor |
H9778 |
| Isosorbide dinitrate powder |
Nitric Oxide Synthase/NO Donor |
I1759 |
| MAHMA NONOate |
Nitric Oxide Synthase/NO Donor |
M1555 |
| Molsidomine |
Nitric Oxide Synthase/NO Donor |
M2901 |
| 3-Morpholinosydnonimine hydrochloride |
Nitric Oxide Synthase/NO Donor |
M5793 |
| 3-Morpholinosydnonimine hydrochloride solid |
Nitric Oxide Synthase/NO Donor |
M184 |
| PAPA NONOate |
Nitric Oxide Synthase/NO Donor |
P8102 |
| 4-Phenyl-3-furoxancarbonitrile |
Nitric Oxide Synthase/NO Donor |
P1726 |
| Proli NONOate |
Nitric Oxide Synthase/NO Donor |
P7977 |
| S150 SPERMINE/NITRIC OXIDE COMPLEX |
Nitric Oxide Synthase/NO Donor |
S150 |
| SE 175 ³97 % |
Nitric Oxide Synthase/NO Donor |
S2189 |
| S-Nitroso-N-acetylpenicillamine ³97% |
Nitric Oxide Synthase/NO Donor |
N3398 |
| S-Nitrosoglutathione ³97% |
Nitric Oxide Synthase/NO Donor |
N4148 |
| Sodium nitroprusside dihydrate |
Nitric Oxide Synthase/NO Donor |
S0501 |
| Streptozocin ³75% as a-anomer ³98% (HPLC) powder |
Nitric Oxide Synthase/NO Donor |
S0130 |
| Sulfo NONOate disodium salt |
Nitric Oxide Synthase/NO Donor |
S8432 |
| Lithium carbamoylphosphate dibasic BioChemika ~85 % (enzymatic) |
Ornithine Carbamoyl-transferase |
21780 |
| Oxaloacetic acid BioChemika ³98.0 % (T) |
Oxaloacetate decarboxylase |
75660 |
| Iodonitrotetrazolium chloride BioChemika for histol. analysis (of oxidases) ³97.0% (AT) |
Oxidase |
58030 |
| S-Benzyl-L-cysteine 4-nitroanilide BioChemika ³98.0 % (NT) |
Oxytocinase |
13340 |
| L-Cystine-di-2-naphthylamide BioChemika ³98.0 % (HPLC) |
Oxytocinase |
30260 |
| P to Z |
Back to top |
| Substrate |
Enzyme |
Product # |
| PRAK Substrate Peptide 1 mg/mL in water ³98 % (HPLC) aqueous solution |
P38 Regulated/Activated Protein Kinase |
P0240 |
| PAK I PEPTIDE SUBSTRATE |
PAK, gamma- (PAK I) |
A-284 |
| Z-L-arginine 7-amido-4-methylcoumarin hydrochloride BioChemika for fluorescence ³90% (TLC) |
Papain |
95938 |
| Na-Benzoyl-L-arginine 4-nitroanilide hydrochloride BioChemika ³98.0 % (enzymatic) |
Papain |
12915 |
| Na-Benzoyl-L-arginine-7-amido-4-methylcoumarin hydrochloride |
Papain |
B7260 |
| Fibrin Blue suitable for substrate for pepsin at low pH (High blanks result at high pH) |
Pepsin |
F5255 |
| Z-Glu-Tyr BioChemika substrate for the det. of pepsin ³99.0 % (HPLC) |
Pepsin |
96150 |
| Phe-Ala-Ala-Phe(4-NO2)-Phe-Val-Leu (4-pyridylmethyl) ester BioChemika ³98.0 % (TLC) |
Pepsin |
77431 |
| N-Benzoyl-L-tyrosyl-4-aminobenzoic acid Sodium salt |
Peptidase |
13136 |
| N-[3-(2-Furyl)acryloyl]-Phe-Gly-Gly BioChemika ³98.0 % (TLC) |
Peptidase |
48176 |
| Naphthol AS-D BioChemika ³99.0 % (HPLC) |
Peptidase |
70489 |
| L-Pyroglutamic acid 2-naphthylamide BioChemika ³99.0 % (TLC) |
Peptidase |
83176 |
| N-Succinyl-Ala-Ala-Pro-Phe p-nitroanilide |
Peptidyl prolyl isomerase |
S7388 |
| N-Oleoylglycine ³98 % |
Peptidylglycine a-amidating enzyme. |
O9762 |
| N-(4-Aminobutyl)-N-ethylisoluminol ³90% |
Peroxidase |
A0156 |
| N-(6-Aminohexyl)-N-ethylisoluminol |
Peroxidase |
A1661 |
| 3-(4-Hydroxyphenyl)propionic acid |
Peroxidase |
H6386 |
| 2,2'-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt BioChemika 10 mg/tablet ABTS |
Peroxidase |
11565 |
| Luminol sodium salt |
Peroxidase |
A4685 |
| Luminol ³97% (HPLC) |
Peroxidase |
A8511 |
| 3-Amino-9-ethylcarbazole ~90% |
Peroxidase |
A5754 |
| 3-Amino-9-ethylcarbazole tablet |
Peroxidase |
A6926 |
| 4-Aminophthalhydrazide monohydrate |
Peroxidase |
A8264 |
| 3-Amino-9-ethylcarbazole BioChemika 20 mg/tablet AEC |
Peroxidase |
03005 |
| 4-Aminoantipyrine reagent grade |
Peroxidase |
A4382 |
| 5-Aminosalicylic acid BioChemika ~100 mg/tablet 5-aminosalicylic acid (enzym.) tablets |
Peroxidase |
09407 |
| 5-Aminosalicylic acid tablet |
Peroxidase |
A6178 |
| 5-Aminosalicylic acid ~99% |
Peroxidase |
A3537 |
| 2,2'-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) Liquid Substrate System |
Peroxidase |
A3219 |
| 2,2'-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt ~98% (TLC) |
Peroxidase |
A1888 |
| 2,2'-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt tablet 10 mg substrate per tablet |
Peroxidase |
A9941 |
| CHEMILUMINESCENT PEROXIDASE SUBSTRATE |
Peroxidase |
CPS-1 |
| 4-Chloro-1-naphthol tablet |
Peroxidase |
C6788 |
| 2-Chloro-5,5-dimethyl-1,3-cyclohexanedione BioChemika suitable as substrate for chloroperoxidase ³98.0 % (AT) |
Peroxidase |
24368 |
| 4-Chloro-1-naphthol BioChemika for analytical purposes ³99.0% (HPLC) |
Peroxidase |
25328 |
| 4-Chloro-1-naphthol BioChemika ³95.0% (HPLC) tablets (~32 mg each) purum |
Peroxidase |
25329 |
| 4-Chloro-1-naphthol purum ³98.0% (T) |
Peroxidase |
25330 |
| 4-Chloro-7-nitrobenzofurazan BioChemika for fluorescence ³99.0% (HPLC) puriss. |
Peroxidase |
25455 |
| 3,3'-Diaminobenzidine (DAB) Enhanced Liquid Substrate System tetrahydrochloride |
Peroxidase |
D3939 |
| 3,3'-Diaminobenzidine (DAB) Enhanced Liquid Substrate System tetrahydrochloride for Membrane Applications |
Peroxidase |
D6815 |
| 3,3'-Diaminobenzidine tetrahydrochloride tablet 10 mg substrate per tablet |
Peroxidase |
D5905 |
| o-Dianisidine dihydrochloride tablet 10 mg substrate per tablet |
Peroxidase |
D9154 |
| o-Dianisidine |
Peroxidase |
D9143 |
| o-Dianisidine dihydrochloride crystalline |
Peroxidase |
D3252 |
| Dicarboxidine dihydrochloride |
Peroxidase |
D5907 |
| Sodium 3,5-dichloro-2-hydroxybenzenesulfonate BioChemika ³98.0 % (TLC) |
Peroxidase |
35867 |
| Dihydrorhodamine 123 BioChemika for fluorescence ³95% (HPLC) |
Peroxidase |
37370 |
| o-Dianisidine dihydrochloride BioChemika ~10 mg/tablet o-dianisidine dihydrochloride |
Peroxidase |
33435 |
| 3-(Dimethylamino)benzoic acid ³97% (HPLC) |
Peroxidase |
D0787 |
| Guaiacol |
Peroxidase |
G5502 |
| Iodonitrotetrazolium chloride BioChemika for histol. analysis (of oxidases) ³97.0% (AT) |
Peroxidase |
58030 |
| Neotetrazolium chloride form BioChemika for microbiology for the detection of reducing fermentation systems ³75% (T) |
Peroxidase |
72150 |
| 3-Nitrotetrazolium blue chloride BioChemika ³75 % (AT) |
Peroxidase |
09972 |
| Nitrotetrazolium blue chloride BioChemika ³88 % (AT) |
Peroxidase |
74030 |
| Nitrotetrazolium blue chloride BioChemika ~10 mg/tablet nitrotetrazolium blue chloride |
Peroxidase |
74031 |
| Nitrotetrazolium blue chloride BioChemika ³98.0 % (calc. based on dry substance, AT) |
Peroxidase |
74032 |
| o-Phenylenediamine BioChemika for fluorescence free base |
Peroxidase |
78411 |
| o-Phenylenediamine dihydrochloride BioChemika for fluorescence |
Peroxidase |
78441 |
| o-Phenylenediamine dihydrochloride BioChemika |
Peroxidase |
78443 |
| o-Phenylenediamine dihydrochloride BioChemika |
Peroxidase |
78446 |
| o-Phenylenediamine dihydrochloride BioChemika |
Peroxidase |
78449 |
| o-Phenylenediamine dihydrochloride crystalline |
Peroxidase |
P1526 |
| o-Phenylenediamine dihydrochloride tablet 60 mg substrate per tablet |
Peroxidase |
P1063 |
| o-Phenylenediamine dihydrochloride tablet 15 mg substrate per tablet |
Peroxidase |
P4664 |
| o-Phenylenediamine dihydrochloride tablet 1 mg substrate per tablet |
Peroxidase |
P6662 |
| o-Phenylenediamine dihydrochloride tablet 2 mg substrate per tablet |
Peroxidase |
P6787 |
| o-Phenylenediamine dihydrochloride tablet 5 mg substrate per tablet |
Peroxidase |
P6912 |
| o-Phenylenediamine dihydrochloride tablet 30 mg substrate per tablet |
Peroxidase |
P8412 |
| o-Phenylenediamine dihydrochloride tablet 4 mg substrate per tablet |
Peroxidase |
P8787 |
| SIGMA FASTä OPD tablet |
Peroxidase |
P9187 |
| o-Phenylenediamine solid |
Peroxidase |
P9029 |
| o-Phenylenediamine tablet 20 mg substrate per tablet |
Peroxidase |
P5412 |
| o-Phenylenediamine dihydrochloride tablet 20 mg substrate per tablet |
Peroxidase |
P7288 |
| o-Phenylenediamine dihydrochloride tablet 3 mg substrate per tablet |
Peroxidase |
P8806 |
| o-Phenylenediamine dihydrochloride tablet 5 mg substrate per tablet |
Peroxidase |
P3804 |
| o-Phenylenediamine dihydrochloride tablet 10 mg substrate per tablet |
Peroxidase |
P8287 |
| Pyrogallol |
Peroxidase |
P0381 |
| Pyrogallol ACS reagent |
Peroxidase |
P2923 |
| SIGMAFAST™ 3,3'-Diaminobenzidine tablets tablet To prepare 1 mL |
Peroxidase |
D4168 |
| SIGMAFAST™ 3,3'-Diaminobenzidine tablets tablet To prepare 5 mL |
Peroxidase |
D4293 |
| SIGMAFAST™ 3,3'-Diaminobenzidine tablets tablet To prepare 15 mL |
Peroxidase |
D4418 |
| SIGMAFAST™ DAB with Metal Enhancer tablet |
Peroxidase |
D0426 |
| 3,3',5,5'-Tetramethylbenzidine dihydrochloride |
Peroxidase |
T8768 |
| 3,3',5,5'-Tetramethylbenzidine (TMB) Liquid Substrate System for ELISA |
Peroxidase |
T0440 |
| 3,3',5,5'-Tetramethylbenzidine (TMB) Liquid Substrate System for Membranes |
Peroxidase |
T0565 |
| 3,3',5,5'-Tetramethylbenzidine Liquid Substrate, Slow Kinetic Form, for ELISA |
Peroxidase |
T4319 |
| 3, 3',5 ,5'-Tetramethylbenzidine Liquid Substrate, Supersensitive, for ELISA |
Peroxidase |
T4444 |
| 3,3',5,5'-Tetramethylbenzidine ³98% (TLC) |
Peroxidase |
T2885 |
| 3,3',5,5'-Tetramethylbenzidine (TMB) Liquid Substrate System |
Peroxidase |
T8665 |
| 3,3',5,5'-Tetramethylbenzidine red liquid substrate system liquid |
Peroxidase |
T1693 |
| 3,3',5,5'-Tetramethylbenzidine dihydrochloride tablet 1 mg substrate per tablet |
Peroxidase |
T3405 |
| 3,3',5,5'-Tetramethylbenzidine tablet 1 mg substrate per tablet |
Peroxidase |
T5525 |
| 3,3',5,5'-Tetramethylbenzidine Liquid Substrate, Super Slow, for ELISA |
Peroxidase |
T5569 |
| Tetranitroblue tetrazolium chloride BioChemika ³85 % (AT) |
Peroxidase |
87961 |
| Tetrazolium Blue Chloride BioChemika indicator for germination, for microbiology ³90% (AT) |
Peroxidase |
88190 |
| Tetrazolium Violet BioChemika ³94 % (AT) |
Peroxidase |
88193 |
| Thiazolyl blue formazan BioChemika |
Peroxidase |
88417 |
| Thiazolyl Blue Tetrazolium Bromide BioChemika ³97.0 % (enzymatic) |
Peroxidase |
88415 |
| trans-5-Phenyl-4-pentenyl hydroperoxide ³90 % Solution in ethanol |
Peroxidase |
P6238 |
| 2,3,5-Triphenyltetrazolium chloride BioChemika ³99.0 % (AT) |
Peroxidase |
93140 |
| Hydrogen peroxide–Urea adduct ³97% |
Peroxidase |
U1753 |
| Hydrogen peroxide–Urea adduct tablet |
Peroxidase |
U8879 |
| Umbelliferyl Arachidonate ethanol solution |
Phopholipase A2 |
U0383 |
| O-(4-Nitrophenylphosphoryl)choline |
Phopholipase C |
N5879 |
| 1-Naphthyl 4-phenylazophenyl phosphate purum ³98.0% (T) |
Phosphodiesterase |
10922 |
| 2-Naphthyl 4-phenylazophenyl phosphate BioChemika ³98.0 % (T) |
Phosphodiesterase |
10923 |
| Thymidine 3'-phosphate 4-nitrophenyl ester sodium salt BioChemika ³80.0 % (HPLC) |
Phosphodiesterase II |
89298 |
| 4-Nitrophenyl phenylphosphonate |
Phosphodiesterase, 5'-Nucleotide |
N2881 |
| Xanthosine 3',5'-cyclic monophosphate ³98 % (HPLC) solid |
Phosphodiesterase, guanosine-3',5'-monophosphate |
X3003 |
| Phospho(enol)pyruvic acid monopotassium salt BioChemika ³98.0 % (PEP-K, enzymatic) |
Phosphoenolpyruvate carboxylase |
79415 |
| Phospho(enol)pyruvic acid monosodium salt monohydrate BioChemika ³98.0 % (NT) |
Phosphoenolpyruvate carboxylase |
79418 |
| Phospho(enol)pyruvic acid tri(cyclohexylammonium) salt BioChemika crystallized ³98.0 % |
Phosphoenolpyruvate carboxylase |
79430 |
| Phospho(enol)pyruvic acid trisodium salt heptahydrate BioChemika ³98.0 % (enzymatic) |
Phosphoenolpyruvate carboxylase |
79435 |
| 6-Phosphogluconic acid barium salt hydrate BioChemika ³95.0 % (TLC) |
Phosphogluconate Dehydrogenase |
79450 |
| 6-Phosphogluconic acid tri(cyclohexylammonium) salt BioChemika ³99.0 % (TLC) |
Phosphogluconate Dehydrogenase |
79458 |
| 6-Phosphogluconic acid trisodium salt BioChemika ³97.0 % (calc. based on dry substance, enzymatic) |
Phosphogluconate Dehydrogenase |
79455 |
| O-(4-Nitrophenylphosphoryl)choline BioChemika ³98.0 % (TLC) |
Phospholipase C |
73739 |
| 1,2-Dipalmitoylphosphatidylinositol 4,5-diphosphate ³98 % solid |
Phospholipase C PI-specific |
P7115 |
| 1,2-Dipalmitoylphosphatidylinositol 3,4-diphosphate ³98 % solid |
Phospholipase C PI-specific |
P6990 |
| Sodium phenyl phosphate dibasic dihydrate BioChemika ³95.0 % (NT) |
Phosphotransferase |
71900 |
| D-Ala-Leu-Lys-7-amido-4-methylcoumarin 95% (HPLC) |
Plasmin |
A8171 |
| Boc-Val-Leu-Lys-7-amido-4-methylcoumarin |
Plasmin |
B1136 |
| Z-Ala-Ala-Lys 4-methoxy-b-naphthylamide formate salt |
Plasmin |
C2147 |
| Gly-Arg-p-nitroanilide dihydrochloride |
Plasmin |
G8148 |
| D-Ile-Phe-Lys p-nitroanilide ³95 % (HPLC) |
Plasmin |
I6886 |
| 4-Nitrophenyl 4-guanidinobenzoate hydrochloride BioChemika ³95.0 % (AT) |
Plasmin |
51015 |
| N-(p-Tosyl)-Gly-Pro-Lys 4-nitroanilide acetate salt |
Plasmin |
T6140 |
| Na-p-Tosyl-L-arginine methyl ester hydrochloride BioChemika for the determination of trypsin ³99.0 % (AT) |
Plasmin |
90170 |
| N-(p-Tosyl)-Gly-Pro-Lys 4-nitroanilide acetate salt BioChemika ³99.0 % (TLC) |
Plasmin |
90178 |
| D-Val-Leu-Lys 4-nitroanilide dihydrochloride |
Plasmin |
V0882 |
| D-Val-Leu-Lys 4-nitroanilide dihydrochloride BioChemika ³98.0 % (HPLC) |
Plasmin |
94680 |
| Porphobilinogen BioChemika (synth.) ³97.0 % (HPLC) |
Porphobilinogen deaminase |
81480 |
| Arg-Glu(EDANS)-[Asn670, Leu671]-Amyloid b/A4 Precursor Protein 770 Fragment 669-674-(DABCYL)Lys-Arg ³96 % (HPLC) solid |
pro-beta-secretase (pro-memapsin 2) |
A2847 |
| Mca-[Asn670, Leu671]-Amyloid b/A4 Precursor Protein 770 fragment 667-675-(Dnp)Lys amide >96 % (HPLC) solid |
pro-beta-secretase (pro-memapsin 2) |
M1063 |
| L-Proline-2-naphthylamide hydrochloride BioChemika ³99.0 % (TLC) |
Proline aminopeptidase |
81742 |
| L-Proline-2-naphthylamide hydrochloride BioChemika ³99.0 % (TLC) |
Proline beta-naphthylamidase |
81742 |
| 2-Aminobenzoyl-Gly-Ala-Ala-Pro-Phe-Tyr(NO2)-Asp trifluoroacetate salt ³90 % (HPLC) |
Proline SpecificEndopeptidase |
A0222 |
| Z-Gly-Pro 7-amido-4-methylcoumarin BioChemika for fluorescence ³97.0% (CHN) |
Prolyl endopeptidase |
96280 |
| Z-Gly-Pro-2-naphthylamide BioChemika for fluorescence ³97.0% (TLC) |
Prolyl endopeptidase |
96283 |
| Z-Gly-Pro-4-nitroanilide BioChemika ³99.0 % (TLC) |
Prolyl endopeptidase |
96286 |
| 1-Naphthyl phosphate disodium salt BioChemika ³99.0 % (HPLC) |
Prostatic acid phosphatase |
71105 |
| N-(3-[2-Furyl]acryloyl)-Ala-Phe amide |
Protease |
F2380 |
| N-(3-[2-Furyl]acryloyl)-Gly-Leu amide |
Protease |
F7383 |
| Albumin, Fluorescein isothiocyanate Conjugate bovine |
Protease |
A9771 |
| Anthranilyl-Asp-Phe-Arg-Leu-Phe-Ala-Phe-nitro-Tyr-Asp ³97% (HPLC) |
Protease |
A0324 |
| Azo dye-impregnated collagen suitable for non-specific protease substraterate of dye release may vary from lot to lot (Dye-impregnated hide powder.) |
Protease |
A4341 |
| Azoalbumin |
Protease |
A2382 |
| Azocasein |
Protease |
A2765 |
| Azo-Casein BioChemika |
Protease |
11610 |
| Azo-Casein sodium salt BioChemika lyophilized |
Protease |
11615 |
| Boric anhydride granulated |
Protease |
11615 |
| Casein, Resorufin-labeled BioChemika |
Protease |
22097 |
| Z-Gly-Gly-Leu p-nitroanilide |
Protease |
C3022 |
| Fibrin Blue suitable for substrate for pepsin at low pH (High blanks result at high pH) |
Protease |
F5255 |
| L-Leucine-7-amido-4-methylcoumarin hydrochloride |
Protease |
L2145 |
| N,N-Dimethylcasein BioChemika for spectrophotometric det. of enzyme activity |
Protease |
39890 |
| N-Succinyl-Gly-Gly-Phe-p-nitroanilide |
Protease B |
S1899 |
| Anthranilyl-Ala-Phe-Ala-Phe-Glu-Val-Phe-nitro-Tyr-Asp ³90% (HPLC) |
Proteases glutamate-specific |
A3567 |
| Z-Leu-Leu-Glu-7-amido-4-methylcoumarin ³95 % solid |
Proteasome |
C0483 |
| Z-Leu-Leu-Leu-7-amido-4-methylcoumarin ³95 % (TLC and HPLC) solid |
Proteasome |
C0608 |
| N-Succinyl-Leu-Leu-Val-Tyr-7-amido-4-trifluoromethylcoumarin ³98 % (TLC) solid |
Proteasome |
S4939 |
| Boc-Leu-Ser-Thr-Arg-7-amido-4-methylcoumarin |
Protein C. |
B4636 |
| [Trp4]-Kemptide ³97% (HPLC) |
Protein Kinase A |
L3523 |
| [Val6, Ala7]-Kemptide ³97% (HPLC) |
Protein Kinase A |
K2877 |
| Arg-Thr-Lys-Arg-Ser-Gly-Ser-Val-Tyr-Glu-Pro-Leu-Lys-Ile ³97 % (HPLC) powder |
Protein Kinase A |
A3317 |
| Kemptide acetate salt ³97% (HPLC) powder |
Protein Kinase A |
K1127 |
| BAD from mouse ³70 % (SDS-PAGE) recombinant, expressed in Escherichia coli buffered aqueous glycerol solution |
Protein Kinase B |
B1682 |
| Myelin Basic Protein Fragment 4-14 ³95% (HPLC) |
Protein Kinase C |
M6913 |
| [Ser25]-Protein Kinase C fragment 19-31 ³97 % (HPLC) |
Protein Kinase C |
P1835 |
| EGF Receptor Fragment 651-658 >95 % (HPLC) |
Protein Kinase C |
E5527 |
| Lys-Arg-Thr-Leu-Arg-Arg trifluoroacetate salt ³97 % (HPLC) |
Protein Kinase C |
L9905 |
| Myelin Basic Protein Fragment 104-118 synthetic lyophilized powder |
Protein Kinase C |
M8184 |
| Neurogranin Fragment 28-43 ³97 % (HPLC) |
Protein Kinase C |
N7279 |
| [pGlu4]-Myelin basic protein fragment 4-14 ³97 % (HPLC) |
Protein Kinase C |
P2186 |
| Pro-Leu-Ser-Arg-Thr-Leu-Ser-Val-Ala-Ala-Lys-Lys ³97% (HPLC) |
Protein Kinase C |
P5307 |
| Val-Arg-Lys-Arg-Thr-Leu-Arg-Arg-Leu ³97 % (HPLC) |
Protein Kinase C |
V2131 |
| Arg-Lys-Arg-Ser-Arg-Ala-Glu lyophilized powder |
Protein kinase G |
A6833 |
| Arg-Lys-Arg-Ser-Arg-Lys-Glu lyophilized powder |
Protein kinase G |
A8457 |
| Protein Tyrosine Phosphatase Substrate Monophosphate ³97% (HPLC) |
Protein Tyrosine Phosphatase |
P6462 |
| L-a-Phosphatidyl-D-myo-inositol 3,4,5-triphosphate, dioctanoyl |
PTEN |
P9953 |
| L-Pyroglutamic acid 7-amido-4-methylcoumarin BioChemika ³99.0 % (HPLC) |
Pyroglutamyl aminopeptidase |
83174 |
| L-Pyroglutamic acid 2-naphthylamide BioChemika ³99.0 % (TLC) |
Pyroglutamyl Peptidase |
83176 |
| Esculin sesquihydrate BioChemika ³98.0 % (HPLC) |
Pyrrolidonyl peptidase |
02350 |
| Phospho(enol)pyruvic acid monopotassium salt BioChemika ³98.0 % (PEP-K, enzymatic) |
Pyruvate kinase |
79415 |
| Phospho(enol)pyruvic acid monosodium salt monohydrate BioChemika ³98.0 % (NT) |
Pyruvate kinase |
79418 |
| Phospho(enol)pyruvic acid tri(cyclohexylammonium) salt BioChemika crystallized ³98.0 % |
Pyruvate kinase |
79430 |
| Phospho(enol)pyruvic acid trisodium salt heptahydrate BioChemika ³98.0 % (enzymatic) |
Pyruvate kinase |
79435 |
| 3-Nitrotetrazolium blue chloride BioChemika ³75 % (AT) |
Quinaldate 4-oxidoreductase |
09972 |
| Nitrotetrazolium blue chloride BioChemika ³88 % (AT) |
Quinaldate 4-oxidoreductase |
74030 |
| Nitrotetrazolium blue chloride BioChemika ~10 mg/tablet nitrotetrazolium blue chloride |
Quinaldate 4-oxidoreductase |
74031 |
| Nitrotetrazolium blue chloride BioChemika ³98.0 % (calc. based on dry substance, AT) |
Quinaldate 4-oxidoreductase |
74032 |
| Tetrazolium Blue Chloride BioChemika indicator for germination, for microbiology ³90% (AT) |
Quinaldate 4-oxidoreductase |
88190 |
| 3-Fluoro-3-deoxy-D-glucose BioChemika ³99.0 % (TLC) |
Reductase |
46865 |
| Renin Substrate 1 |
Renin |
R8276 |
| N-Benzoyl-Phe-Val-Arg-p-nitroanilide hydrochloride BioChemika ³99.0 % (TLC) |
Reptilase |
13042 |
| Ribonucleic acid from baker's yeast (S. cerevisiae) BioChemika ~18 A260/mg |
RNase |
83847 |
| NFAT 1 from mouse ³90% (SDS-PAGE) recombinant, expressed in Escherichia coli N-terminal 6-His tagged Truncated sequence amino acids 1-297 buffered aqueous glycerol solution |
SAPK2a/p38a |
N9659 |
| TNPAL-Sphingomyelin 1 mg/mL in chloroform: methanol (2:1 v/v) |
Sphingomyelinase |
T1014 |
| Na-p-Tosyl-L-arginine methyl ester hydrochloride BioChemika for the determination of trypsin ³99.0 % (AT) |
Subtilisin |
90170 |
| 2-(2'-Benzothiazolyl)-5-styryl-3-(4'-phthalhydrazidyl)tetrazolium chloride 90-95% |
Succinic dehydrogenase |
B8131 |
| Iodonitrotetrazolium chloride BioChemika for histol. analysis (of oxidases) ³97.0% (AT) |
Succinic Dehydrogenase |
58030 |
| 4-[3-(4-Iodophenyl)-2-(2,4-dinitrophenyl)-2H-5-tetrazolio]-1,3-benzenedisulfonate sodium salt BioChemika ³90 % (HPLC) |
Succinic Dehydrogenase |
90482 |
| 3-Nitrotetrazolium blue chloride BioChemika ³75 % (AT) |
Succinic Dehydrogenase |
09972 |
| Nitrotetrazolium blue chloride BioChemika ³88 % (AT) |
Succinic Dehydrogenase |
74030 |
| Nitrotetrazolium blue chloride BioChemika ~10 mg/tablet nitrotetrazolium blue chloride |
Succinic Dehydrogenase |
74031 |
| Nitrotetrazolium blue chloride BioChemika ³98.0 % (calc. based on dry substance, AT) |
Succinic Dehydrogenase |
74032 |
| Tetranitroblue tetrazolium chloride BioChemika ³85 % (AT) |
Succinic Dehydrogenase |
87961 |
| Tetrazolium Blue Chloride BioChemika indicator for germination, for microbiology ³90% (AT) |
Succinic Dehydrogenase |
88190 |
| 2,3,5-Triphenyltetrazolium chloride BioChemika ³99.0 % (AT) |
Succinic Dehydrogenase |
93140 |
| 1,3,5-Triphenyltetrazolium formazan BioChemika ³97.0 % (UV) |
Succinic Dehydrogenase |
93145 |
| 5-Bromo-4-chloro-3-indolyl sulfate potassium salt BioChemika ³99.0 % (T) |
Sulfatase |
16673 |
| 5-Bromo-6-chloro-3-indolyl sulfate potassium salt BioChemika ³99.0 % (HPLC) |
Sulfatase |
16674 |
| 4-Nitrocatechol sulfate dipotassium salt monohydrate BioChemika ³97.0 % (T) |
Sulfatase |
56020 |
| 4-Methylumbelliferyl sulfate potassium salt BioChemika for fluorescence ³99.0% (HPLC) |
Sulfatase |
69610 |
| Phenolphthalein disulfate tripotassium salt BioChemika ³99.0 % (TLC) |
Sulfatase |
77635 |
| 3-Nitrotetrazolium blue chloride BioChemika ³75 % (AT) |
Superoxid dismutase |
09972 |
| Nitrotetrazolium blue chloride BioChemika ³88 % (AT) |
Superoxid dismutase |
74030 |
| Nitrotetrazolium blue chloride BioChemika ~10 mg/tablet nitrotetrazolium blue chloride |
Superoxid dismutase |
74031 |
| Nitrotetrazolium blue chloride BioChemika ³98.0 % (calc. based on dry substance, AT) |
Superoxid dismutase |
74032 |
| Tetrazolium Blue Chloride BioChemika indicator for germination, for microbiology ³90% (AT) |
Superoxid dismutase |
88190 |
| Methyl gallate purum antioxidant ³98.0% (GC) |
Tannase |
48690 |
| 7-Methoxycoumarin-4-acetyl-[Asn670, Leu671]-Amyloid b/A4 Precursor Protein 770 Fragment 667-676-(2,4-dinitrophenyl)Lys-Arg-Arg amide ³90 % (HPLC) |
Thimet oligopeptidase (TOP) |
A1472 |
| 7-Methoxycoumarin-4-acetyl-[Asn670,Leu671]-Amyloid b/A4 Precursor Protein 770 (667-676)-(2,4-dinitrophenyl)Lys-Arg-Arg Amide ³96 % (HPLC) |
Thimet oligopeptidase (TOP) |
A1347 |
| 3-Nitrotetrazolium blue chloride BioChemika ³75 % (AT) |
Thiophene-2-carbonyl-CoA monooxygenase |
09972 |
| Nitrotetrazolium blue chloride BioChemika ³88 % (AT) |
Thiophene-2-carbonyl-CoA monooxygenase |
74030 |
| Nitrotetrazolium blue chloride BioChemika ~10 mg/tablet nitrotetrazolium blue chloride |
Thiophene-2-carbonyl-CoA monooxygenase |
74031 |
| Nitrotetrazolium blue chloride BioChemika ³98.0 % (calc. based on dry substance, AT) |
Thiophene-2-carbonyl-CoA monooxygenase |
74032 |
| Tetrazolium Blue Chloride BioChemika indicator for germination, for microbiology ³90% (AT) |
Thiophene-2-carbonyl-CoA monooxygenase |
88190 |
| N-Benzoyl-Phe-Val-Arg-p-nitroanilide hydrochloride BioChemika ³99.0 % (TLC) |
Thrombin |
13042 |
| N-Benzoyl-Phe-Val-Arg 4-methoxy-b-naphthylamide hydrochloride |
Thrombin |
B1260 |
| N-Benzoyl-Phe-Val-Arg-p-nitroanilide hydrochloride |
Thrombin |
B7632 |
| Boc-b-benzyl-Asp-Pro-Arg-7-amido-4-methylcoumarin hydrochloride ~95% |
Thrombin |
B4028 |
| Boc-Val-Pro-Arg-7-amido-4-methylcoumarin hydrochloride |
Thrombin |
B9385 |
| 4-Methylumbelliferyl 4-guanidinobenzoate hydrochloride hydrate crystalline |
Thrombin |
M4149 |
| 4-Methylumbelliferyl 4-guanidinobenzoate hydrochloride hydrate BioChemika for fluorescence |
Thrombin |
51010 |
| 4-Nitrophenyl 4-guanidinobenzoate hydrochloride BioChemika ³95.0 % (AT) |
Thrombin |
51015 |
| Sar-Pro-Arg p-nitroanilide dihydrochloride |
Thrombin |
S9009 |
| Thrombin generation chromogenic substrate ³90% (TLC) solid |
Thrombin |
T3068 |
| N-p-Tosyl-Gly-Pro-Arg 7-amido-4-methylcoumarin hydrochloride |
Thrombin |
T0273 |
| N-(p-Tosyl)-Gly-Pro-Arg p-nitroanilide acetate salt |
Thrombin |
T1637 |
| Na-p-Tosyl-L-arginine methyl ester hydrochloride BioChemika for the determination of trypsin ³99.0 % (AT) |
Thrombin |
90170 |
| N-(p-Tosyl)-Gly-Pro-Arg p-nitroanilide acetate salt BioChemika ³98.0 % (TLC) |
Thrombin |
90175 |
| N-Acetyl-L-phenylalanyl-3,5-diiodo-L-tyrosine BioChemika ³97.0 % (CHN) |
Thyroid-hormone transaminase |
01408 |
| pGlu-Gly-Arg-7-amido-4-trifluoromethylcoumarin ³95 % (HPLC) lyophilized powder |
Tissue Plasinogen Activator |
P5615 |
| Tissue plasminogen activator chromogenic substrate ³95 % (HPLC) solid |
Tissue Plasinogen Activator |
T2943 |
| D-Val-Leu-Lys 7-amido-4-methylcoumarin ³99 % (HPLC) lyophilized powder |
Tissue Plasinogen Activator |
V3138 |
| D-Val-Leu-Lys 7-amido-4-trifluoromethylcoumarin lyophilized powder |
Tissue Plasinogen Activator |
V3013 |
| D-Val-Leu-Lys 4-methoxy-2-naphthylamide solid |
Tissue Plasinogen Activator |
V3263 |
| D-Val-Phe-Lys p-Nitroanilide dihydrochloride ³97% (HPLC) |
Tissue Plasinogen Activator |
V6884 |
| Pefachrome® tPA BioChemika ³95.0 % (HPLC) |
Tissue Plasminogen Activator |
94174 |
| 3-Amino-2,3-dihydrobenzoic acid hydrochloride purum ³98.0% (calc. based on dry substance, T) |
Transaminase, gamma- |
07710 |
| AQUANAL®-plus ammonium (NH4) 0.2-8.0 mg/l refill pack for 37400 |
Triosephosphate isomerase |
37440 |
| Dihydroxyacetone phosphate dilithium salt BioChemika ~95 % (enzymatic) |
Triosephosphate isomerase |
37440 |
| Dihydroxyacetone phosphate dilithium salt technical ~90 % (enzymatic) |
Triosephosphate isomerase |
37442 |
| AQUANAL®-plus chromium (Cr) 0.005-0.1 mg/l refill pack for 37402 |
Triosephosphate isomerase |
37442 |
| Z-L-arginine 7-amido-4-methylcoumarin hydrochloride BioChemika for fluorescence ³90% (TLC) |
Trypsin |
95938 |
| Azo-Casein BioChemika |
Trypsin |
11610 |
| Azo-Casein sodium salt BioChemika lyophilized |
Trypsin |
11615 |
| Boric anhydride granulated |
Trypsin |
11615 |
| Na-Benzoyl-L-arginine ethyl ester hydrochloride |
Trypsin |
B4500 |
| Na-Benzoyl-DL-arginine 4-nitroanilide hydrochloride ³98% |
Trypsin |
B4875 |
| Na-Benzoyl-L-arginine ethyl ester hydrochloride BioChemika ³99.0% (AT) |
Trypsin |
12880 |
| Na-Benzoyl-L-arginine 4-nitroanilide hydrochloride BioChemika ³98.0 % (enzymatic) |
Trypsin |
12915 |
| Cobalt puriss. ³99.9% wire |
Trypsin |
12915 |
| Na-Benzoyl-L-arginyl-4-amino-benzoic acid hydrochloride BioChemika ³99.0 % (sum of enantiomers, TLC) |
Trypsin |
12925 |
| Na-Benzoyl-DL-arginine b-naphthylamide hydrochloride BioChemika ³97.0 % (AT) |
Trypsin |
12910 |
| Na-Benzoyl-DL-arginine 4-nitroanilide hydrochloride BioChemika ³98.0% (AT) |
Trypsin |
12920 |
| Na-Benzoyl-L-isoleucyl-L-glutamyl-glycyl-L-arginine-4-nitroanilide BioChemika ³97.0 % (HPLC) |
Trypsin |
87528 |
| N-Benzoyl-Phe-Val-Arg-p-nitroanilide hydrochloride BioChemika ³99.0 % (TLC) |
Trypsin |
13042 |
| N-Benzoyl-Phe-Val-Arg-p-nitroanilide hydrochloride |
Trypsin |
B7632 |
| Boc-Gln-Ala-Arg-7-amido-4-methylcoumarin hydrochloride ~95% |
Trypsin |
B4153 |
| Casein, Resorufin-labeled BioChemika |
Trypsin |
22097 |
| Z-Gly-Gly-Arg b-naphthylamide hydrochloride ~98% |
Trypsin |
C7784 |
| Hide–Remazol Brilliant Blue R |
Trypsin |
H6268 |
| Hide–Remazol Brilliant Blue R BioChemika |
Trypsin |
89435 |
| 4-Methylumbelliferyl 4-guanidinobenzoate hydrochloride hydrate crystalline |
Trypsin |
M4149 |
| 4-Methylumbelliferyl 4-guanidinobenzoate hydrochloride hydrate BioChemika for fluorescence |
Trypsin |
51010 |
| N,N-Dimethylcasein BioChemika for spectrophotometric det. of enzyme activity |
Trypsin |
39890 |
| Na-Benzoyl-L-arginine-7-amido-4-methylcoumarin hydrochloride |
Trypsin |
B7260 |
| Na-Benzoyl-L-arginine 4-methoxy-b-naphthylamide hydrochloride |
Trypsin |
B0885 |
| Na-Benzoyl-L-arginine methyl ester bicarbonate salt |
Trypsin |
B4256 |
| Na-Benzoyl-L-arginine 4-nitroanilide hydrochloride |
Trypsin |
B3133 |
| Na-Benzoyl-L-arginine 4-nitroanilide hydrochloride |
Trypsin |
B3279 |
| Na-Benzoyl-DL-arginine b-naphthylamide hydrochloride |
Trypsin |
B4750 |
| Z-Arg-Arg-Arg 4-methoxy-b-naphthylamide |
Trypsin |
C5645 |
| 4-Nitrophenyl 4-guanidinobenzoate hydrochloride BioChemika ³95.0 % (AT) |
Trypsin |
51015 |
| Pefachrome® Fxa BioChemika ³95.0 % (HPLC) |
Trypsin |
77739 |
| Pefachrome® tPA BioChemika ³95.0 % (HPLC) |
Trypsin |
94174 |
| Pefachrome® Try BioChemika ³95.0 % (HPLC) |
Trypsin |
77723 |
| Na-p-Tosyl-L-arginine methyl ester hydrochloride BioChemika for the determination of trypsin ³99.0 % (AT) |
Trypsin |
90170 |
| N-Acetyl-Lys-Pro-Arg-7-amido-4-trifluoromethylcoumarin >95 % (HPLC) solid |
Tryptase |
C6608 |
| Tyramine BioChemika for fluorescence ³99.0% (TLC) |
Tyramine dehydrogenase |
93810 |
| Nedd8 human ³95 % (SDS-PAGE) recombinant, expressed in Escherichia coli buffered aqueous solution |
Ubc12 |
N8529 |
| Nedd8 Precursor human ³95 % (SDS-PAGE) recombinant, expressed in Escherichia coli buffered aqueous solution |
Ubc12 |
N2030 |
| SUMO-1 human ³95 % (SDS-PAGE) recombinant, expressed in Escherichia coli |
UbcH9 |
S0314 |
| SUMO-1 human ³95 % (SDS-PAGE) recombinant, expressed in Escherichia coli (GST-tagged) |
UbcH9 |
S0439 |
| Urea BioChemika Ultra ³99.5% (T) |
Urea carboxylase |
51458 |
| 3-(1-Acetylacetonylazo)phthalhydrazide BioChemika ³97.0 % (HPLC) |
Urease |
17751 |
| Urea BioChemika Ultra ³99.5% (T) |
Urease |
51458 |
| Na-p-Tosyl-L-arginine methyl ester hydrochloride BioChemika for the determination of trypsin ³99.0 % (AT) |
Urinary esterase |
90170 |
| N-Benzoyl-Val-Gly-Arg p-nitroanilide hydrochloride |
Urokinase |
B4758 |
| Z-Gly-Gly-Arg 7-amido-4-methylcoumarin hydrochloride |
Urokinase |
C9396 |
| Z-Gly-Gly-Arg 4-methoxy-b-naphthylamide hydrochloride |
Urokinase |
C5770 |
| N-Glutaryl-Gly-Arg-7-amido-4-methylcoumarin hydrochloride |
Urokinase |
G2386 |
| Gly-Arg-p-nitroanilide dihydrochloride |
Urokinase |
G8148 |
| Pefachrome® uPA BioChemika ³95.0 % (HPLC) |
Urokinase |
77713 |
| pGlu-Gly-Arg-7-amido-4-trifluoromethylcoumarin ³95 % (HPLC) lyophilized powder |
Urokinase |
P5615 |
| Iodonitrotetrazolium chloride BioChemika for histol. analysis (of oxidases) ³97.0% (AT) |
Xanthine dehydrogenase |
58030 |
| 4-O-Methyl-D-glucurono-D-xylan dyed with Remazol brilliant blue R BioChemika 15 % as dye |
Xylanase, endo-1,4-beta- |
66960 |
| Ostazin Brilliant Red H-3B hydroxyethyl cellulose |
Xylanase, endo-1,4-beta- |
O6879 |
| 4-Methylumbelliferyl-b-D-xylopyranoside |
Xylosidase, beta- |
M7008 |
| 4-Methylumbelliferyl-b-D-xylopyranoside BioChemika ³99.0 % (TLC) |
Xylosidase, beta- |
69617 |
| 4-Nitrophenyl b-D-xylopyranoside ³98% |
Xylosidase, beta- |
N2132 |
| 4-Nitrophenyl b-D-xylopyranoside BioChemika ³99.0 % (TLC) |
Xylosidase, beta- |
73750 |
| LAT human ~50 % (glutathione-agarose chromatography) recombinant, expressed in Escherichia coli buffered aqueous glycerol solution |
ZAP 70 Kinase |
L9413 |