Sigma-Aldrich
Life Science > Metabolomics > Enzyme Explorer > Substrate Index
Enzyme Explorer
Substrate Index
TO FIND SUBSTRATES FOR AN ENZYME...
  1. Search alphabetically for the 12915 enzyme in the table below
  2. You will find substrates and Product Numbers on the same line
  3. Click on the Product # to access product listing and related information including:
    Product Information Sheets, C of A and MSDS
  A to C
  D to N
  P to Z
A to C
Substrate Enzyme Product # 
Acetylthiocholine iodide BioChemika Acetylcholinesterase 01480
5-Bromo-4-chloro-3-indolyl N-acetyl-b-D-glucosaminide ³98 % Acetylglucosaminidase, beta-N- B3041
4-Methylumbelliferyl-7-(6-sulfo-2-acetamido-2-deoxy-b-D-glucopyranoside) sodium salt ~98% (TLC) Acetylglucosaminidase, beta-N- M0662
4-Methylumbelliferyl N-acetyl-b-D-glucosaminide Acetylglucosaminidase, beta-N- M2133
4-Methylumbelliferyl N-acetyl-a-D-glucosaminide Acetylglucosaminidase, beta-N- M9881
4-Nitrophenyl N-acetyl-b-D-glucosaminide 98-100% Acetylglucosaminidase, beta-N- N9376
4-Nitrophenyl N-acetyl-b-D-galactosaminide ³98% Acetylhexosaminidase, beta-N- N9003
sn-Glycerol 3-phosphate bis(cyclohexylammonium) salt BioChemika ³95 % (GC) Acid Phosphatase 49990
Indoxyl acetate BioChemika ³97.0% (TLC) Acid Phosphatase 57420
Naphthol AS-BI phosphate BioChemika for histology ³98.0% (HPLC) Acid Phosphatase 70491
Naphthol AS-BS phosphate BioChemika for histology Acid Phosphatase 70483
Naphthol AS-E phosphate BioChemika for histology Acid Phosphatase 70485
Naphthol AS-MX phosphate BioChemika for histology ³99.0% (HPLC) Acid Phosphatase 70476
Naphthol AS-MX phosphate disodium salt nonahydrate BioChemika for histology ³99.0% (TLC) Acid Phosphatase 70495
Naphthol AS phosphate BioChemika for histology Acid Phosphatase 70471
Naphthol AS-TR phosphate BioChemika for histology ³98.0% (HPLC) Acid Phosphatase 70478
Naphthol AS-TR phosphate disodium salt BioChemika for histology ~4 mg/tablet naphthol AS-TR-phosphate tablets Acid Phosphatase 70467
NBT-BCIP solution BioChemika suitable as substrate for alkaline phosphatase in dot blots Acid Phosphatase 72091
4-Nitrophenyl phosphate bis(cyclohexylammonium) salt BioChemika for the determination of phosphatase ~95 % (enzymatic) Acid Phosphatase 73736
4-Nitrophenyl phosphate bis[tris(hydroxymethyl)aminomethane] salt BioChemika for the determination of phosphatase ³97.0 % (enzymatic) Acid Phosphatase 73737
Phenolphthalein bisphosphate pyridine salt BioChemika ³95 % (T) Acid Phosphatase 77630
Thymolphthalein monophosphate disodium salt hydrate BioChemika ~75% (NT) Acid Phosphatase 89362
Tris(4-nitrophenyl) phosphate BioChemika ³95 % (HPLC) Acid Phosphatase 93380
N-Acetyl-Met-Ala ³97 % (HPLC) Acylamino acid releasing enzyme A1450
Indoxyl acetate BioChemika ³97.0% (TLC) Acylase 57420
Nebularine BioChemika ³98.0 % (HPLC) Adenosine deaminase 82590
N-Benzoyl-Pro-Phe-Arg-p-nitroanilide hydrochloride Agkistrodon B2133
3-Fluoro-3-deoxy-D-glucose BioChemika ³99.0 % (TLC) Aldolase reductase 46865
Alkaline Phosphatase Yellow (pNPP) Liquid Substrate System for ELISA liquid Alkaline Phosphatase A3469
BCIP/NBT-Blue Liquid Substate System for Membranes Alkaline Phosphatase B3804
BCIP/NBT-Purple Liquid Substate System for Membranes Alkaline Phosphatase B3679
BCIP/NBT Liquid Substrate System Alkaline Phosphatase B1911
Bis(4-nitrophenyl) phosphate hydrate purum ³97.0% (dried material, T) Alkaline Phosphatase 15125
SIGMAFAST™ BCIP/NBT tablet Alkaline Phosphatase B5655
5-Bromo-4-chloro-3-indolyl phosphate disodium salt Alkaline Phosphatase B6149
5-Bromo-4-chloro-3-indolyl phosphate dipotassium salt Alkaline Phosphatase B6274
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt tablet Alkaline Phosphatase B0274
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt ³98% Alkaline Phosphatase B8503
5-Bromo-6-chloro-3-indolyl phosphate p-toluidine salt ~97% (HPLC) Alkaline Phosphatase B5667
5-Bromo-4-chloro-3-indolyl phosphate disodium salt BioChemika ³98.0 % (HPLC) Alkaline Phosphatase 16668
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt BioChemika ³90 % (HPLC) Alkaline Phosphatase 16670
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt BioChemika ~25 mg/tablet 5-bromo-4-chloro-3-indolyl phosphate p-toluidine salt tablets Alkaline Phosphatase 16676
5-Bromo-6-chloro-3-indolyl phosphate p-toluidine salt BioChemika ³95.0 % (HPLC) Alkaline Phosphatase 16672
CDP-Star® Chemiluminescent Substrate 0.25 mM in water Alkaline Phosphatase C0712
Ammonium cerium(IV) sulfate dihydrate ³99 % Alkaline Phosphatase 31173
2-Deoxy-a-D-ribose 1-phosphate bis(cyclohexylammonium) salt BioChemika ³95 % (TLC) Alkaline Phosphatase 31173
Fast Red RC tablet 10 mg substrate per tablet Alkaline Phosphatase F5146
Fast Red TR Salt hemi(zinc chloride) salt Alkaline Phosphatase F8764
Indoxyl phosphate disodium salt crystalline Alkaline Phosphatase I5505
Indoxyl phosphate disodium salt BioChemika ³95.0 % (TLC) Alkaline Phosphatase 57440
D-Luciferin 6'-O-phosphate trisodium salt BioChemika ³99.0 % (TLC) Alkaline Phosphatase 62657
D-Luciferin 6'-O-phosphate trisodium salt lyophilized powder Alkaline Phosphatase L9402
4-Methylumbelliferyl phosphate BioChemika for fluorescence ³99.0% (HPLC) Alkaline Phosphatase 69607
4-Methylumbelliferyl phosphate dilithium salt Alkaline Phosphatase M9044
4-Methylumbelliferyl phosphate disodium salt Alkaline Phosphatase M8168
4-Methylumbelliferyl phosphate disodium salt BioChemika for fluorescence ³99.0% (HPLC) Alkaline Phosphatase 69605
4-Methylumbelliferyl phosphate Alkaline Phosphatase M8883
4-Methylumbelliferyl phosphate (4-MUP) Liquid Substrate System liquid Alkaline Phosphatase M3168
Naphthol AS-BI phosphate disodium salt Alkaline Phosphatase N2250
Naphthol AS-BI phosphate BioChemika for histology ³95.0% (HPLC) Alkaline Phosphatase 70482
Naphthol AS-BI phosphate disodium salt BioChemika for histology ³99.0% (TLC) Alkaline Phosphatase 70494
Naphthol AS-E phosphate BioChemika for histology Alkaline Phosphatase 70485
Naphthol AS-MX phosphate Alkaline Phosphatase N4875
Naphthol AS-MX phosphate BioChemika for histology ³99.0% (HPLC) Alkaline Phosphatase 70476
Naphthol AS-MX phosphate disodium salt nonahydrate BioChemika for histology ³99.0% (TLC) Alkaline Phosphatase 70495
Naphthol AS phosphate BioChemika for histology Alkaline Phosphatase 70471
Naphthol AS-TR phosphate disodium salt Alkaline Phosphatase N6125
Naphthol AS-TR phosphate BioChemika for histology ³98.0% (HPLC) Alkaline Phosphatase 70478
Naphthol AS-TR phosphate disodium salt BioChemika for histology ~4 mg/tablet naphthol AS-TR-phosphate tablets Alkaline Phosphatase 70467
1-Naphthyl phosphate sodium salt ³98% (titration) powder Alkaline Phosphatase N7000
2-Naphthyl phosphate sodium salt Alkaline Phosphatase N7375
1-Naphthyl phosphate disodium salt Alkaline Phosphatase N7255
1-Naphthyl phosphate disodium salt BioChemika ³99.0 % (HPLC) Alkaline Phosphatase 71105
2-Naphthyl phosphate sodium salt BioChemika ³99.0 % (HPLC) Alkaline Phosphatase 71099
2-Naphthyl phosphate sodium salt BioChemika ³98.0 % (HPLC) Alkaline Phosphatase 71100
1-Naphthyl phosphate monosodium salt monohydrate BioChemika ³98.0 % (HPLC) Alkaline Phosphatase 71090
NBT-BCIP solution BioChemika suitable as substrate for alkaline phosphatase in dot blots Alkaline Phosphatase 72091
Nitro Blue Tetrazolium tablet Alkaline Phosphatase N5514
4-Nitrophenyl phosphate bis(cyclohexylammonium) salt Alkaline Phosphatase N3129
4-Nitrophenyl phosphate bis(cyclohexylammonium) salt BioChemika for the determination of phosphatase ~95 % (enzymatic) Alkaline Phosphatase 73736
4-Nitrophenyl phosphate bis[tris(hydroxymethyl)aminomethane] salt BioChemika for the determination of phosphatase ³97.0 % (enzymatic) Alkaline Phosphatase 73737
4-Nitrophenyl phosphate di(2-amino-2-ethyl-1,3-propanediol) salt Alkaline Phosphatase N6260
4-Nitrophenyl phosphate disodium salt hexahydrate tablet Alkaline Phosphatase N2640
4-Nitrophenyl phosphate disodium salt hexahydrate tablet Alkaline Phosphatase N2765
4-Nitrophenyl phosphate disodium salt hexahydrate tablet Alkaline Phosphatase N9389
4-Nitrophenyl phosphate disodium salt BioChemika purum tablets 20 mg/tablet substrate (remainder binding agent) Alkaline Phosphatase 73724
4-Nitrophenyl phosphate disodium salt BioChemika ³99.0 % (5 mg substrate per tablet, rest binding agent, TLC) tablets Alkaline Phosphatase 73730
4-Nitrophenyl phosphate disodium salt hexahydrate BioChemika for enzyme immunoassay ³99.0 % (enzymatic) Alkaline Phosphatase 71768
4-Nitrophenyl phosphate disodium salt hexahydrate BioChemika ³97.0 % (NT) Alkaline Phosphatase 71770
4-Nitrophenyl phosphate di(tris) salt Alkaline Phosphatase N3254
p-Nitrophenyl Phosphate Liquid Substrate System liquid Alkaline Phosphatase N7653
4-Nitrophenyl phosphate magnesium salt Alkaline Phosphatase N2507
3-Nitrotetrazolium blue chloride BioChemika ³75 % (AT) Alkaline Phosphatase 09972
Nitrotetrazolium blue chloride BioChemika ³88 % (AT) Alkaline Phosphatase 74030
Nitrotetrazolium blue chloride BioChemika ~10 mg/tablet nitrotetrazolium blue chloride Alkaline Phosphatase 74031
Nitrotetrazolium blue chloride BioChemika ³98.0 % (calc. based on dry substance, AT) Alkaline Phosphatase 74032
Phenolphthalein bisphosphate pyridine salt BioChemika ³95 % (T) Alkaline Phosphatase 77630
Phenolphthalein monophosphate bis(cyclohexylammonium) salt BioChemika ³98.0 % (HPLC) Alkaline Phosphatase 77639
Phenolphthalein monophosphate bis(cyclohexylammonium) salt ~90% powder Alkaline Phosphatase P5758
3-Phenylumbelliferyl phosphate hemipyridine salt BioChemika for fluorescence ³85% (HPLC) Alkaline Phosphatase 79281
SIGMAFAST™ Fast Red TR/Naphthol AS-MX Tablets tablet To prepare 10 mL Alkaline Phosphatase F4523
SIGMAFAST™ Fast Red TR/Naphthol AS-MX Tablets tablet To prepare 1 mL Alkaline Phosphatase F4648
SIGMAFASTä p-Nitrophenyl phosphate Tablets tablet To prepare 5 mL Alkaline Phosphatase N1891
SIGMAFASTä p-Nitrophenyl phosphate Tablets tablet To prepare 20 mL Alkaline Phosphatase N2770
5-(Tetradecanoylamino)fluorescein BioChemika for fluorescence ³95% (HPLC) Alkaline Phosphatase 87180
Tetrazolium Blue Chloride BioChemika indicator for germination, for microbiology ³90% (AT) Alkaline Phosphatase 88190
Thymolphthalein monophosphate disodium salt hydrate BioChemika ~75% (NT) Alkaline Phosphatase 89362
Tris(4-nitrophenyl) phosphate BioChemika ³95 % (HPLC) Alkaline Phosphatase 93380
Na-Benzoyl-DL-arginine 4-nitroanilide hydrochloride BioChemika ³98.0% (AT) Amidase 12920
3-Amino-2,3-dihydrobenzoic acid hydrochloride purum ³98.0% (calc. based on dry substance, T) Aminobutyric acid transaminase, gamma- 07710
1-Aminocyclopropanecarboxylic acid BioChemika ³98.0 % (NT) Aminocyclopropanecarboxyl oxidase 07626
Succinyl coenzyme A disodium salt BioChemika ³90 % (HPLC) Aminolevulinate synthase, 5- 85985
5-Aminolevulinic acid hydrochloride BioChemika ³97.0 % (AT) aminolevulinic acid dehydratase, delta- 08340
L-Alanine 7-amido-4-methylcoumarin trifluoroacetate salt BioChemika for fluorescence ³99.0% (TLC) Aminopeptidase 05198
L-Alanine 4-nitroanilide hydrochloride BioChemika ³99.0 % (TLC) Aminopeptidase 05215
Z-L-arginine 7-amido-4-methylcoumarin hydrochloride BioChemika for fluorescence ³90% (TLC) Aminopeptidase 95938
L-Citrulline 7-amido-4-methylcoumarin hydrobromide BioChemika ³98.0 % (sum of enantiomers, HPLC) Aminopeptidase 27525
Z-Gly-Pro 7-amido-4-methylcoumarin BioChemika for fluorescence ³97.0% (CHN) Aminopeptidase 96280
L-Methionine 7-amido-4-methylcoumarin trifluoroacetate salt BioChemika ³99.0 % (sum of enantiomers, HPLC) Aminopeptidase 64367
L-Proline-2-naphthylamide hydrochloride BioChemika ³99.0 % (TLC) Aminopeptidase 81742
L-Pyroglutamic acid 7-amido-4-methylcoumarin BioChemika ³99.0 % (HPLC) Aminopeptidase 83174
Ammonium thiocyanate puriss. ³99% (AT) Aminopeptidase 11235
L-Aspartic acid b-(7-amido-4-methylcoumarin) BioChemika ³98.0 % (TLC) Aminopeptidase 11235
Fmoc-L-glutamic acid 1-(7-amido-4-methylcoumarin) BioChemika ³98.0 % (HPLC) Aminopeptidase A 02243
L-Glutamic acid 1-(4-nitroanilide) BioChemika ³98.0 % (T) Aminopeptidase A 49622
L-Leucine 7-amido-4-methyl coumarin hydrochloride BioChemika for fluorescence Aminopeptidase B 61888
N-Acetyl-Arg-Gly-Phe-Phe-Pro 4-methoxy-2-naphthylamide >90 % (TLC) solid Aminopeptidase M C7108
L-Leucine p-nitroanilide BioChemika ³99.0 % (NT) Aminopeptidase M 61910
L-Phenylalanine 7-amido-4-methylcoumarin trifluoroacetate salt BioChemika for fluorescence ³99.0% (sum of enantiomers, HPLC) Aminopeptidase M 78089
4-Nitrophenyl a-D-maltopentaoside ³98% Amylase N1519
Starch Azure Insoluble corn starch covalently linked with Remazol Brilliant Blue R Amylase S7776
Starch Azure Potato starch covalently linked with Remazol Brilliant Blue R Amylase S7629
Amylopectin Azure Amylase, alpha- A4640
Amylose–Remazol Brilliant Blue R Amylase, alpha- A3508
Amylose–Remazol Brilliant Blue R BioChemika Amylase, alpha- 10140
2-Chloro-4-nitrophenyl-b-D-maltoheptaoside BioChemika ~80 % (HPLC) Amylase, alpha- 25590
2-Chloro-4-nitrophenyl-b-D-maltotrioside BioChemika ³96.0 % (HPLC) Amylase, alpha- 25591
4-Nitrophenyl a-D-maltoside Amylase, alpha- N5885
Starch dyed with Remazol brilliant blue R BioChemika Amylase, alpha- 85641
Z-Val-Lys-Met 7-amido-4-methylcoumarin Amyloid A4-generating enzymes C2681
N-[3-(2-Furyl)acryloyl]-Phe-Gly-Gly Angiotensin Converting Enzyme F7131
Hippuryl-Gly-Gly Angiotensin Converting Enzyme H1763
Hippuryl-His-Leu acetate salt Angiotensin Converting Enzyme H4884
N-Hippuryl-His-Leu tetrahydrate Angiotensin Converting Enzyme H1635
N-Hippuryl-His-Leu tetrahydrate BioChemika ³99.0 % (HPLC) Angiotensin-converting enzyme 53285
4-Nitrophenyl a-L-arabinofuranoside BioChemika ³99.0 % (TLC) Arabinofuranosidase III 73616
4-Nitrophenyl a-L-arabinofuranoside ³98% (TLC) Arabinofuranosidase III, alpha-L- N3641
4-Methylumbelliferyl a-L-arabinopyranoside Arabinosidase M3167
5-Hydroxy-L-tryptophan solid Aromatic amino acid decarboxylase, L- H9772
Tryptamine hydrochloride BioChemika ³98.0 % (AT) Arylamine N-methyltransferase 93650
6-Chloro-3-indolyl phosphate p-toluidine salt BioChemika ³98.0 % (HPLC) Aryl-sulfatase 24908
4-Nitrocatechol sulfate dipotassium salt monohydrate BioChemika ³97.0 % (T) Arylsulfatases A and B 56020
Na-Acetyl-L-asparagine BioChemika ³98.0 % (T) Asparaginase II 00915
Oxaloacetic acid BioChemika ³98.0 % (T) Aspartate transaminase 75660
Adenosine 5'-triphosphate P3-[1-(2-nitrophenyl)ethyl ester] disodium salt BioChemika ³95% (HPLC) ATPase 02070
N-Methoxysuccinyl-Phe-Leu-Phe-7-amido-4-trifluoromethylcoumarin solid ATP-dependent protease M4813
N-Methoxysuccinyl-Phe-Leu-Phe 4-methoxy-2-naphthylamide ³90 % solid ATP-dependent protease M4938
Amyloid b/A4 Precursor Protein 770 Fragment 667-676 ³96 % (HPLC) solid beta-Secretase A3222
[Asn670, Leu671]-Amyloid b/A4 Precursor Protein 770 Fragment 667-676 trifluoroacetate salt ³95 % (HPLC) solid beta-Secretase A1972
DABCYL-[Asn670, Leu671]-Amyloid b/A4 Precursor Protein 770 fragment 661-675-EDANS trifluoroacetate salt ³97 % (HPLC) solid beta-Secretase A4972
DABCYL-[Asn670, Leu671]-Amyloid b/A4 Precursor Protein 770 Fragment 669-674-EDANS ammonium salt ³95 % (HPLC) solid beta-Secretase A5097
7-Methoxycoumarin-4-acetyl-[Asn670, Leu671]-Amyloid b/A4 Precursor Protein 770 Fragment 669-674-(2,4-dinitrophenyl)Lys trifluoroacetate salt ³96 % (HPLC) beta-Secretase A1597
7-Methoxycoumarin-4-acetyl-[Asn670, Leu671]-Amyloid b/A4 Precursor Protein 770 Fragment 667-674-(2,4-dinitrophenol)Dap ³97 % (HPLC) beta-Secretase A1222
7-Methoxycoumarin-4-acetyl-[Asn670, Leu671]-Amyloid b/A4 Precursor Protein 770 Fragment 667-676-(2,4-dinitrophenyl)Lys-Arg-Arg amide ³90 % (HPLC) beta-Secretase A1472
7-Methoxycoumarin-4-acetyl-[Asn670,Leu671]-Amyloid b/A4 Precursor Protein 770 (667-676)-(2,4-dinitrophenyl)Lys-Arg-Arg Amide ³96 % (HPLC) beta-Secretase A1347
[Val671]-Amyloid b/A4 Precursor Protein 770 Fragment 667-676 trifluoroacetate salt ³95 % (HPLC) solid beta-Secretase A2722
N-(+)-Biotinyl-4-aminobenzoic acid BioChemika ³98.0 % (HPLC) Biotinidase 14408
Zinc puriss. ³99.99% granulated Biotinidase 14404
Biotin 4-amidobenzoic acid sodium salt BioChemika ³99.0 % (TLC) Biotinidase 14404
Zinc purum ³99% powder Biotinidase 14409
Biocytin BioChemika ³97.0 % (HPLC) Biotinidase 14409
Na-Benzoyl-L-arginine 4-nitroanilide hydrochloride BioChemika ³98.0 % (enzymatic) Bromelain 12915
Z-L-Lys-ONp hydrochloride Bromelain C3637
Z-Lys-ONp hydrochloride purum ³98.0% (AT) Bromelain 96895
4-Methylumbelliferyl butyrate BioChemika for fluorescence ³95% (HPCE) Butyrate esterase 19362
4-Nitrophenyl butyrate BioChemika ³98.0 % (HPLC) Butyrylcholinesterase 19368
Autocamtide 2 trifluoroacetate salt ³97 % (HPLC) Calmodulin-dependent protein kinase II A0591
Syntide 2 ³97% (HPLC) Calmodulin-dependent protein kinase II S2525
Syntide 2 BioChemika ³99.0 % (HPLC) Calmodulin-dependent protein kinase II 86221
Calcium/calmodulin-dependent protein kinase II-d fragment 345-358 Calmodulin-dependent protein kinase IV C0843
L-Methionine 7-amido-4-methylcoumarin trifluoroacetate salt BioChemika ³99.0 % (sum of enantiomers, HPLC) Calpain 64367
N-Succinyl-Leu-Leu-Val-Tyr-7-Amido-4-Methylcoumarin ³90% (HPLC) Calpain S6510
N-Succinyl-Leu-Tyr-7-amido-4-methylcoumarin Calpain S1153
Acetyl-L-phenylalanyl-L-3-thiaphenylalanine BioChemika ³98.0 % (HPLC) Carboxypeptidase A 29223
Hippuryl-Lys Carboxypeptidase B H6750
Acetyl-Lys-D-Ala-D-Ala ³95 % (HPLC) Carboxypeptidase G A6950
N-(3-[2-Furyl]acryloyl)-Ala-Lys powder Carboxypeptidase N F5882
Na,Ne-Diacetyl-Lys-D-Ala-D-Ala Carboxypeptidase, penicillin-sensitive D-alanine D9904
Na,Ne-Diacetyl-Lys-D-Ala-D-lactic acid Carboxypeptidase, penicillin-sensitive D-alanine D2279
N-(3-[2-Furyl]acryloyl)-Phe-Phe Carboxypeptidase, Serum F7133
Hippuryl-L-phenylalanine BioChemika ³97.0 % (TLC) CarboxypeptidaseA 53284
Hippuryl-L-phenylalanine CarboxypeptidaseA H6875
L-Carnosine BioChemika ³99.0 % (NT) Carnosinase 22030
Casein Kinase II substrate ³97% (HPLC) Casein Kinase II C2460
N-Acetyl-Val-Ala-Asp-7-amido-4-trifluoromethylcoumarin >98 % (HPLC) solid Caspase (general) A0723
N-Acetyl-Val-Ala-Asp p-nitroanilide >98 % (HPLC) solid Caspase (general) A0848
L-Asp-Asp-Rhodamine trifluoroacetate salt >98 % solid Caspase (general) D5189
N-Acetyl-Trp-Glu-His-Asp-7-amido-4-trifluoromethylcoumarin trifluoroacetate salt ³97 % (HPLC) powder Caspase 1 A6720
N-Acetyl-Trp-Glu-His-Asp-p-nitroanilide ³98 % (HPLC) powder Caspase 1 A6845
N-Acetyl-Tyr-Val-Ala-Asp p-nitroanilide ³97% (HPLC) powder Caspase 1 A3831
N-Acetyl-Trp-Glu-His-Asp-7-amido-4-methylcoumarin ³97 % (HPLC) powder Caspase 1 A0216
N-Acetyl-Tyr-Val-Ala-Asp-7-amido-4-trifluoromethylcoumarin ³95 % (HPLC) powder Caspase 1 A9965
7-Methoxycoumarin-4-acetyl-Tyr-Val-Ala-Asp-Ala-Pro-(2,4-dinitrophenyl)Lys ³97 % (HPLC) Caspase 1 M5295
N-Acetyl-Tyr-Val-Ala-Asp-7-amido-4-methylcoumarin ³99% (TLC) powder Caspase 1 A2452
N-Acetyl-Val-Asp-Val-Ala-Asp-7-amido-4-trifluoromethylcoumarin ³97 % (HPLC) powder Caspase 2 A5345
N-Acetyl-Val-Asp-Val-Ala-Asp-p-nitroanilide ³97 % (HPLC) powder Caspase 2 A5470
N-Acetyl-Val-Asp-Val-Ala-Asp-7-amido-4-trifluoromethylcoumarin ³97 % (HPLC) powder Caspase 2 A5345
N-Acetyl-Val-Asp-Val-Ala-Asp-p-nitroanilide ³97 % (HPLC) powder Caspase 2 A5470
N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-trifluoromethylcoumarin ³90 % (HPLC) powder Caspase 3 A0466
N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-trifluoromethylcoumarin ³90 % (HPLC) powder Caspase 3 A0466
N-Acetyl-Asp-Gln-Met-Asp-7-amido-4-trifluoromethylcoumarin ³90 % (HPLC) powder Caspase 3 A5595
N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-methylcoumarin ³97% (HPLC) powder Caspase 3 A1086
N-Acetyl-Asp-Glu-Val-Asp p-nitroanilide ³97% (HPLC) powder Caspase 3 A2559
N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-trifluoromethylcoumarin ³90 % (HPLC) powder Caspase 3 A0466
N-Acetyl-Asp-Glu-Val-Asp p-nitroanilide ³97% (HPLC) powder Caspase 3 A2559
DFF45/ICAD human >85 % (SDS-PAGE) recombinant, expressed in Escherichia coli solution Caspase 3 D3815
N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-methylcoumarin ³97% (HPLC) powder Caspase 3 A1086
N-Acetyl-Asp-Glu-Val-Asp p-nitroanilide ³97% (HPLC) powder Caspase 3 A2559
N-Acetyl-Asp-Gln-Met-Asp-7-amido-4-trifluoromethylcoumarin ³90 % (HPLC) powder Caspase 3 A5595
N-Acetyl-Leu-Glu-Val-Asp-7-amido-4-trifluoromethylcoumarin ³95 % (HPLC) solid Caspase 4 A2099
N-Acetyl-Tyr-Val-Ala-Asp p-nitroanilide ³97% (HPLC) powder Caspase 4 A3831
N-Acetyl-Tyr-Val-Ala-Asp-7-amido-4-methylcoumarin ³99% (TLC) powder Caspase 4 A2452
N-Acetyl-Tyr-Val-Ala-Asp-7-amido-4-trifluoromethylcoumarin ³95 % (HPLC) powder Caspase 4 A9965
N-Acetyl-Trp-Glu-His-Asp-7-amido-4-trifluoromethylcoumarin trifluoroacetate salt ³97 % (HPLC) powder Caspase 5 A6720
N-Acetyl-Trp-Glu-His-Asp-p-nitroanilide ³98 % (HPLC) powder Caspase 5 A6845
N-Acetyl-Trp-Glu-His-Asp-p-nitroanilide ³98 % (HPLC) powder Caspase 5 A6845
N-Acetyl-Trp-Glu-His-Asp-p-nitroanilide ³98 % (HPLC) powder Caspase 5 A6845
N-Acetyl-Val-Glu-His-Asp-7-amido-4-trifluoromethylcoumarin trifluoroacetate salt ³98 % (HPLC) solid Caspase 6 A2097
N-Acetyl-Val-Glu-Ile-Asp-7-amido-4-methylcoumarin ³90 % (HPLC) powder Caspase 6 A0341
N-Acetyl-Val-Glu-Ile-Asp-7-amido-4-trifluoromethylcoumarin ³97 % (HPLC) powder Caspase 6 A5095
N-Acetyl-Val-Glu-Ile-Asp-7-amido-4-methylcoumarin ³90 % (HPLC) powder Caspase 6 A0341
N-Acetyl-Val-Glu-Ile-Asp-p-nitroanilide ³97 % (HPLC) powder Caspase 6 A5220
CASPASE-6 ASSAY KIT, FLUORIMETRIC Caspase 6 CASP-6-F
N-Acetyl-Val-Glu-Ile-Asp-7-amido-4-methylcoumarin ³90 % (HPLC) powder Caspase 6 A0341
N-Acetyl-Val-Glu-Ile-Asp-7-amido-4-trifluoromethylcoumarin ³97 % (HPLC) powder Caspase 6 A5095
N-Acetyl-Val-Glu-Ile-Asp-p-nitroanilide ³97 % (HPLC) powder Caspase 6 A5220
N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-trifluoromethylcoumarin ³90 % (HPLC) powder Caspase 7 A0466
N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-trifluoromethylcoumarin ³90 % (HPLC) powder Caspase 7 A0466
N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-methylcoumarin ³97% (HPLC) powder Caspase 7 A1086
N-Acetyl-Asp-Glu-Val-Asp p-nitroanilide ³97% (HPLC) powder Caspase 7 A2559
N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-methylcoumarin ³97% (HPLC) powder Caspase 7 A1086
N-Acetyl-Asp-Glu-Val-Asp p-nitroanilide ³97% (HPLC) powder Caspase 7 A2559
N-Acetyl-Ala-Glu-Val-Asp-7-amido-4-trifluoromethylcoumarin >96 % (HPLC) solid Caspase 8 A0973
N-Acetyl-Ala-Glu-Val-Asp-7-amido-4-trifluoromethylcoumarin >96 % (HPLC) solid Caspase 8 A0973
N-Acetyl-Ile-Glu-Thr-Asp-p-nitroanilide ³95 % Caspase 8 A9968
N-Acetyl-Leu-Glu-Thr-Asp-7-Amido-4-trifluoromethylcoumarin ³97 % (HPLC) powder Caspase 8 A6095
N-Acetyl-Ile-Glu-Thr-Asp-7-Amido-4-methylcoumarin ³97 % (HPLC) powder Caspase 8 A4188
CASPASE-8 ASSAY KIT, COLORIMETRIC Caspase 8 CASP-8-C
CASPASE-8 ASSAY KIT, FLUORIMETRIC Caspase 8 CASP-8-F
Z-Ile-Glu-Thr-Asp 7-amido-4-trifluoromethylcoumarin ³95 % (HPLC) Caspase 8 C5599
N-Acetyl-Ile-Glu-Thr-Asp-7-Amido-4-methylcoumarin ³97 % (HPLC) powder Caspase 8 A4188
N-Acetyl-Leu-Glu-Thr-Asp-7-Amido-4-trifluoromethylcoumarin ³97 % (HPLC) powder Caspase 8 A6095
N-Acetyl-Ile-Glu-Thr-Asp-p-nitroanilide ³95 % Caspase 8 A9968
Z-Ile-Glu-Thr-Asp 7-amido-4-trifluoromethylcoumarin ³95 % (HPLC) Caspase 8 C5599
N-Acetyl-Leu-Glu-His-Asp-7-amido-4-trifluoromethylcoumarin ³95 % (HPLC) powder Caspase 9 A5845
N-Acetyl-Val-Glu-His-Asp-7-amido-4-trifluoromethylcoumarin trifluoroacetate salt ³98 % (HPLC) solid Caspase 9 A2097
N-Acetyl-Leu-Glu-His-Asp-7-amido-4-trifluoromethylcoumarin ³95 % (HPLC) powder Caspase 9 A5845
N-Acetyl-Leu-Glu-His-Asp-p-nitroanilide ³95 % (HPLC) powder Caspase 9 A5970
Hydrogen peroxide solution puriss. p.a. ACS reagent ³30% (RT) Catalase 95300
Z-Phe-Ala puriss. ³99.0% (T) Cathepsin A 97029
Z-Ala-Arg-Arg 4-methoxy-b-naphthylamide acetate salt Cathepsin B C8536
Z-Phe-Arg 4-methoxy-b-naphthylamide hydrochloride Cathepsin B C3282
Z-Val-Lys-Lys-Arg-4-methoxy-b-naphthylamide trihydrochloride Cathepsin B C2772
Hippuryl-Arg Cathepsin B H2508
Na-Benzoyl-L-arginine 4-methoxy-b-naphthylamide hydrochloride Cathepsin B B0885
Z-Arg-Arg-7-amido-4-methylcoumarin hydrochloride Cathepsin B C5429
Z-Arg-Arg 4-methoxy-b-naphthylamide acetate salt Cathepsin B C5520
N-Succinyl-Leu-Leu-Val-Tyr-7-Amido-4-Methylcoumarin ³90% (HPLC) Cathepsin B S6510
Gly-Phe b-naphthylamide crystalline Cathepsin C G4255
Gly-Phe b-naphthylamide crystalline Cathepsin C G9512
Gly-Phe p-nitroanilide Cathepsin C G0142
Ser-Tyr b-naphthylamide powder Cathepsin C S5628
N-Acetyl-Arg-Gly-Phe-Phe-Pro-7-amido-4-methylcoumarin solid Cathepsin D C6358
N-Acetyl-Arg-Gly-Phe-Phe-Pro-7-amido-4-trifluoromethylcoumarin >95 % (HPLC) solid Cathepsin D C6983
N-Acetyl-Arg-Gly-Phe-Phe-Pro 4-methoxy-2-naphthylamide >90 % (TLC) solid Cathepsin D C7108
Boc-Phe-Ala-Ala-p-Nitro-Phe-Phe-Val-Leu 4-hydroxymethylpyridine ester Cathepsin D B1644
7-Methoxycoumarin-4-Acetyl-Gly-Lys-Pro-Ile-Leu-Phe-Phe-Arg-Leu-Lys(DNP)-D- Arg-amide >97 % (HPLC) solid Cathepsin D M0938
7-Methoxycoumarin-4-Acetyl-Gly-Lys-Pro-Ile-Leu-Phe-Phe-Arg-Leu-Lys(DNP)-D- Arg-amide >97 % (HPLC) solid Cathepsin E M0938
N-Methoxysuccinyl-Ala-Ala-Pro-Met p-nitroanilide >95 % (HPLC) Cathepsin G M7771
N-Succinyl-Ala-Ala-Pro-Phe p-nitroanilide Cathepsin G S7388
N-Succinyl-Ala-Ala-Pro-Phe thiobenzyl ester Cathepsin G S6518
N-Succinyl-Gly-Gly-Phe-7-amido-4-trifluoromethylcoumarin ³98 % (TLC) solid Cathepsin G S4689
N-Succinyl-Gly-Gly-Phe-4-methoxy-2-naphthylamide ³98 % (TLC) solid Cathepsin G S4814
N-Succinyl-Phe-Leu-Phe thiobenzyl ester Cathepsin G S6643
Z-Phe-Arg 4-methoxy-b-naphthylamide hydrochloride Cathepsin L C3282
N-Succinyl-Ala-Ala-Pro-Phe p-nitroanilide Cathepsin, Leukocyte S7388
Cellotetraose BioChemika ~95 % (HPLC) Cellulase 22164
Cellulose Azure Cellulase C1052
2-Chloro-4-nitrophenyl-b-D-cellobioside BioChemika chromophoric enzyme substrate ³98.0 % (HPCE) Cellulase 25586
4-Methylumbelliferyl b-D-cellobioside Cellulase M6018
2-Nitrophenyl b-D-cellobioside Cellulase N4764
4-Nitrophenyl b-D-cellobioside Cellulase N5759
4-Nitrophenyl b-D-cellobioside BioChemika ³99.0 % (HPLC) Cellulase 73617
Chitin azure Chitinase C3020
4-Methylumbelliferyl-N,N'-diacetyl-b-D-chitobioside monohydrate BioChemika for fluorescence ³99.0% (HPLC) Chitinase 69589
4-Nitrophenyl N,N'-diacetyl-b-D-chitobioside Chitinase N6133
4-Nitrophenyl N,N'-diacetyl-b-D-chitobioside BioChemika ³99.0 % (TLC) Chitinase 73623
CHKtide, CHK1 Substrate Peptide liquid CHK1 C7734
2-Chloro-5,5-dimethyl-1,3-cyclohexanedione BioChemika suitable as substrate for chloroperoxidase ³98.0 % (AT) Chloroperoxidase 24368
7-Acetoxy-1-methylquinolinium iodide BioChemika for fluorescence ³98.0% (HPCE) Cholinesterase 00871
Benzoylcholine chloride BioChemika ³98.0 % (AT) Cholinesterase 12950
Butyrylcholine iodide BioChemika ³99.0 % (AT) Cholinesterase 20780
b-Carbonaphthoxycholine iodide Cholinesterase C7125
2,6-Dichlorophenolindophenyl acetate BioChemika ³97.0% (CHN) Cholinesterase 36190
Pencycuron PESTANAL®, analytical standard Cholinesterase 36190
Indoxyl acetate BioChemika ³97.0% (TLC) Cholinesterase 57420
4-Nitrophenyl butyrate Cholinesterase
Resorufin butyrate BioChemika for fluorescence ³95% (TLC) Cholinesterase 83637
Succinylcholine chloride dihydrate BioChemika ³99.0% (AT) Cholinesterase 85980
N-Succinyl-Ala-Ala-Pro-Phe p-nitroanilide Chymotrypsin, alpha S7388
N-Succinyl-Ala-Ala-Pro-Phe-7-amido-4-methylcoumarin Chymotrypsin, alpha S9761
N-Acetyl-L-tyrosine ethyl ester monohydrate BioChemika ³99.0 % (sum of enantiomers, HPLC) Chymotrypsin, alpha- 01530
Ala-Ala-Phe-7-amido-4-methylcoumarin Chymotrypsin, alpha- A3401
Ala-Ala-Val-Ala p-nitroanilide Chymotrypsin, alpha- A9273
N-Benzoyl-L-tyrosine ethyl ester BioChemika ³98.0% (NT) Chymotrypsin, alpha- 13110
Magnesium according to Grignard ³99.5 % turnings Chymotrypsin, alpha- 13110
N-Benzoyl-L-tyrosine ethyl ester Chymotrypsin, alpha- B6125
N-Benzoyl-L-tyrosine p-nitroanilide Chymotrypsin, alpha- B6760
N-Benzoyl-L-tyrosine p-amidobenzoic acid sodium salt BioChemika lyophilized ³98.0 % (sum of enantiomers, HPLC) Chymotrypsin, alpha- 13135
Glutaryl-Ala-Ala-Phe-4-methoxy-b-naphthylamide Chymotrypsin, alpha- G3769
N-Glutaryl-Gly-Gly-Phe-b-naphthylamide Chymotrypsin, alpha- G4636
Glutaryl-L-phenylalanine 7-amido-4-methylcoumarin BioChemika for fluorescence ³98.0% (HPLC) Chymotrypsin, alpha- 49737
N-Glutaryl-L-phenylalanine p-nitroanilide BioChemika ³98.0 % (HPLC) Chymotrypsin, alpha- 49738
N-(N-Glutaryl-L-phenylalanyl)-2-aminoacridone BioChemika for fluorescence ³98.0% (HPLC) Chymotrypsin, alpha- 49742
4-Methylumbelliferyl p-trimethylammoniocinnamate chloride powder Chymotrypsin, alpha- M4507
N-Succinyl-Ala-Ala-Pro-Phe 7-amido-4-trifluoromethylcoumarin ³99 % (TLC) solid Chymotrypsin, alpha- S5189
N-Succinyl-Ala-Ala-Pro-Phe 4-methoxy-2-naphthylamide ³98 % (TLC) solid Chymotrypsin, alpha- S5314
N-Succinyl-Ala-Ala-Pro-Phe p-nitroanilide BioChemika ³99.0 % (HPLC) Chymotrypsin, alpha- 85977
N-Succinyl-Gly-Gly-Phe-p-nitroanilide Chymotrypsin, alpha- S1899
N-Succinyl-L-phenylalanine-p-nitroanilide Chymotrypsin, alpha- S2628
N-(N-Succinyl-L-phenylalanyl)-2-aminoacridone BioChemika for fluorescence ³98.0% (HPCE) Chymotrypsin, alpha- 85992
Oxaloacetic acid BioChemika ³98.0 % (T) Citrate synthase 75660
c-Jun human 40-50 % (SDS-PAGE) recombinant, expressed in Escherichia coli truncated human c-Jun sequence-GST fusion protein 1-169-GST, soluble c-Jun Kinase C5859
c-Jun Kinase substrate from rat >90 % (SDS-PAGE) recombinant, expressed in Escherichia coli buffered aqueous solution c-Jun Kinase C3233
Succinyl coenzyme A disodium salt BioChemika ³90 % (HPLC) CoA ligase, succinate- 85985
N-[3-(2-Furyl)acryloyl]-Leu-Gly-Pro-Ala Collagenase F5135
N-(2,4-Dinitrophenyl)-Pro-Leu-Gly-Cys(Me)-His-Ala-D-Arg amide ³95 % (HPLC) Collagenase D0296
N-[3-(2-Furyl)acryloyl]-Leu-Gly-Pro-Ala BioChemika ³99.0% (HPLC) Collagenase 48173
Z-Gly-Pro-Gly-Gly-Pro-Ala Collagenase C8008
H-Lys(Z)-OH 98 % Collagenase C8008
Z-Gly-Pro-Leu-Gly-Pro Collagenase C0627
COLLAGEN-FLUORESCEIN Collagenase C4486
Collagen Type I-FITC Conjugate bovine skin ~1 mg/mL solution Collagenase C4361
Collagen-Fluorescein bovine Collagenase C5858
Collagenase Chromophore-Substrate (for quantitative Collagenase-Determination) BioChemika Collagenase 27668
Collagenase Chromophore-Substrate (for quantitative Collagenase-Determination) Components A + B BioChemika Collagenase 27669
Collagenase Chromophore-Substrate (for quantitative Collagenase-Determination) Components A + B BioChemika Collagenase 27671
Collagenase-Substrate (for quantitative Collagenase-Determination) Components A + B BioChemika Collagenase 27670
Collagenase-Substrate (for quantitative Collagenase-Determination) Components A + B BioChemika Collagenase 27672
Collagenase-Substrate (for quantitative Collagenase-Determination) BioChemika Collagenase 27673
Collagenase-Substrate (for quantitative Collagenase-Determination) Component B BioChemika Collagenase 27674
Glycolic acid solution ~57 % Collagenase 27674
7-Methoxycoumarin-4-acetyl-Pro-(cyclohexyl)-Ala-Gly-Nva-His-Ala-L-(2,4-dinitrop henyl)diaminoproponic acid solid Collagenase-3 M3314
Boc-Leu-Gly-Arg-7-amido-4-methylcoumarin acetate salt Convertase B4511
Urea BioChemika Ultra ³99.5% (T) Cyanamide hydratase 51458
Resorufin benzyl ether Cytochrome P450 B1532
Resorufin methyl ether Cytochrome P450 M1544
Resorufin ethyl ether Cytochrome P450 E3763
7-Benzyloxy-4-trifluoromethylcoumarin 99 % (by 1H-NMR) solid Cytochrome P450 1A2 B5057
7-Ethoxycoumarin BioChemika for fluorescence ³99.0% (HPLC) Cytochrome P450 2B6 02644
7-Methoxy-4-trifluoromethylcoumarin 99 % (by 1H-NMR) Cytochrome P450 2C9 M8189
7-Benzyloxyquinoline 99% (by 1H-NMR) solid Cytochrome P450 3A4 B5182
7-Benzyloxy-4-trifluoromethylcoumarin 99 % (by 1H-NMR) solid Cytochrome P450 3A4 B5057
3-Nitrotetrazolium blue chloride BioChemika ³75 % (AT) Cytokinin dehydrogenase 09972
Nitrotetrazolium blue chloride BioChemika ³88 % (AT) Cytokinin dehydrogenase 74030
Nitrotetrazolium blue chloride BioChemika ~10 mg/tablet nitrotetrazolium blue chloride Cytokinin dehydrogenase 74031
Nitrotetrazolium blue chloride BioChemika ³98.0 % (calc. based on dry substance, AT) Cytokinin dehydrogenase 74032
Tetrazolium Blue Chloride BioChemika indicator for germination, for microbiology ³90% (AT) Cytokinin dehydrogenase 88190
N-Acetyl-Gly(t-butyl)-Gly(t-butyl)-Asn(Methyl)2-Ala-7-amido-4-methylcoumari n ³97 % (HPLC) solid Cytomegalovirus (HCMV) protease A2347
2-Aminobenzoyl-Gly(t-butyl)-Asn(methyl)2-Ala-Ser-Ser-Arg-Leu-Tyr(NO2)-Tyr-Arg trifluoroacetate salt >94 % (HPLC) Cytomegalovirus (HCMV) protease A9846
Resorufin acetate BioChemika for fluorescence ³98.0% (TLC) Cytosolic Aldehyde Dehydrogenase (Esterase Activity) 83636
D to N Back to top
Substrate Enzyme Product # 
Resorufin pentyl ether BioChemika for fluorescence ³98.0% (HPLC) Dealkylase 77105
Neotetrazolium chloride form BioChemika for microbiology for the detection of reducing fermentation systems ³75% (T) Dehydrogenase 72150
Thiazolyl blue formazan BioChemika Dehydrogenase 88417
Thiazolyl Blue Tetrazolium Bromide BioChemika ³97.0 % (enzymatic) Dehydrogenase 88415
2,3,5-Triphenyltetrazolium chloride BioChemika ³99.0 % (AT) Dehydrogenase 93140
1,3,5-Triphenyltetrazolium formazan BioChemika ³97.0 % (UV) Dehydrogenase 93145
N-Acetyl-L-phenylalanyl-3,5-diiodo-L-tyrosine BioChemika ³97.0 % (CHN) Diiodotyrosine transaminase 01408
Lys-Ala-7-amido-4-methylcoumarin dihydrochloride Dipeptidyl aminopeptidase L8139
Gly-Arg 4-methoxy-b-naphthylamide dihydrochloride Dipeptidyl aminopeptidase I G1013
Lys-Ala 4-methoxy-b-naphthylamide dihydrochloride Dipeptidyl aminopeptidase II L2270
Arg-Arg b-naphthylamide trihydrochloride Dipeptidyl aminopeptidase III A3261
Arg-Arg 4-methoxy-b-naphthylamide trihydrochloride Dipeptidyl aminopeptidase III A9398
Arg-Pro p-nitroanilide acetate salt Dipeptidyl aminopeptidase IV A1204
Gly-Pro 4-methoxy-b-naphthylamide hydrochloride powder Dipeptidyl aminopeptidase IV G9262
Gly-Pro 4-methoxy-b-naphthylamide Dipeptidyl aminopeptidase IV G9137
Gly-Pro p-nitroanilide hydrochloride >99% (TLC) Dipeptidyl peptidase IV G0513
Gly-Pro-7-amido-4-methylcoumarin hydrobromide Dipeptidyl peptidase IV. G2761
Gly-Pro p-nitroanilide p-toluenesulfonate salt ~99% (TLC) Dipeptidyl peptidase IV. G2901
2,3-Diphospho-D-glyceric acid penta(cyclohexylammonium) salt BioChemika ³97.0 % (HPCE) Diphosphoglycerate-independent phosphoglycerate mutase 43380
Tyramine BioChemika for fluorescence ³99.0% (TLC) Dopamine beta-hydroxylase 93810
N-(Methoxysuccinyl)-Ala-Ala-Pro-Val 4-nitroanilide BioChemika ³97.0 % (HPLC) Elastase 65352
N-Succinyl-Ala-Ala-Ala-p-nitroanilide Elastase S4760
N-Succinyl-Ala-Ala-Ala-p-nitroanilide BioChemika ³99.0 % (enzymatic) Elastase 85975
Boc-D-Phe-Ala-Nle-p-nitroanilide Elastase, Hunan Granulocyte B8397
Boc-Ala-Pro-Nva p-chlorothiobenzyl ester Elastase, leukocyte B2017
N-Methoxysuccinyl-Ala-Ala-Pro-Val-7-amido-4-methylcoumarin Elastase, leukocyte M9771
N-(Methoxysuccinyl)-Ala-Ala-Pro-Val 4-nitroanilide Elastase, leukocyte M4765
N-Methoxysuccinyl-Ala-Ala-Pro-Val-7-amido-4-trifluoromethylcoumarin solid Elastase, leukocyte M4563
N-Methoxysuccinyl-Ala-Ala-Pro-Val-7-amido-4-methylcoumarin Elastase, pancreatic M9771
7-Methoxycoumarin-4-acetyl [Ala7-(2,4-Dinitrophenyl)Lys9]-Bradykinin trifluoroacetate salt >94 % (HPLC) Endothelin-converting enzyme-1 B5181
N-Dansyl-D-Ala-Gly-p-nitro-Phe-Gly ~95% (HPLC) Enkephalinase D2155
Glutaryl-Ala-Ala-Phe-4-methoxy-b-naphthylamide Enkephalinase G3769
Gly-Asp-Asp-Asp-Asp-Lys-b-naphthylamide ³95% (HPLC) Enterokinase G5261
(2S,3S)-trans-3-Phenyl-2-oxiranylmethyl 4-nitrophenyl carbonate BioChemika ³98.0 % (NMR) Epoxy-Hydrolase 04088
2',7'-Dichlorofluorescin diacetate BioChemika for fluorescence ³90% (HPLC) Esterase 35847
8-Acetoxy-N,N,N',N',N'',N''-hexamethylpyrene-1,3,6-trisulfonamide BioChemika for fluorescence ³98.0% (TLC) Esterase 52612
8-Acetoxypyrene-1,3,6-trisulfonic acid trisodium salt BioChemika for fluorescence ³98.0% (TLC) Esterase 00879
N-Acetyl-L-tyrosine ethyl ester monohydrate BioChemika ³99.0 % (sum of enantiomers, HPLC) Esterase 01530
3-(2-Benzoxazolyl)umbelliferyl acetate BioChemika for fluorescence ³98.0% (HPLC) Esterase 12832
5-Bromo-4-chloro-3-indolyl acetate Esterase B4377
5-Bromo-4-chloroindoxyl 1,3-diacetate BioChemika ³99.0 % (HPLC) Esterase 16675
5-Bromoindoxyl acetate BioChemika ³98.0 % (AT) Esterase 17400
8-Butyryloxypyrene-1,3,6-trisulfonic acid trisodium salt BioChemika for fluorescence ³95.0% (TLC) Esterase 20800
Paraoxon-ethyl ~90% oil Esterase D9286
Fluorescein dibutyrate BioChemika for fluorescence ³98.0% (HPCE) Esterase 46942
Fluorescein dilaurate BioChemika for fluorescence ³97.0% (HPCE) Esterase 46943
Indoxyl acetate Esterase I3500
4-Methylumbelliferyl acetate BioChemika for fluorescence ³97.0% (TLC) Esterase 46002
4-Methylumbelliferyl butyrate BioChemika for fluorescence ³95% (HPCE) Esterase 19362
Na-Benzoyl-L-arginine ethyl ester hydrochloride BioChemika ³99.0% (AT) Esterase 12880
Naphthol AS acetate BioChemika for histology ³99.0% (HPLC) Esterase 70465
Naphthol AS-D acetate BioChemika for histology ³95.0% (HPLC) Esterase 70472
Naphthol AS-D acetate BioChemika ³99.0 % (HPLC) Esterase 70488
Naphthol AS chloroacetate BioChemika for histology ³98.0% (HPLC) Esterase 70470
Naphthol AS-D chloroacetate BioChemika for histology ³95.0% (HPLC) Esterase 70468
Naphthol AS-D chloroacetate Esterase N0758
Naphthol AS-E acetate BioChemika for histology ³96.0% (HPLC) Esterase 70484
Naphthol AS-MX acetate BioChemika for histology ~90% (HPLC) Esterase 70475
1-Naphthyl acetate BioChemika ³99.0 % (HPLC) Esterase 46010
Primisulfuron-methyl PESTANAL®, analytical standard Esterase 46010
2-Naphthyl acetate BioChemika substrate for the det. of esterase ³99.0 % (GC) Esterase 46020
1-Naphthyl acetate Esterase N8505
1-Naphthyl butyrate Esterase N8000
1-Naphthyl butyrate BioChemika ³99.0 % (GC) Esterase 19363
4-Nitrophenyl butyrate Esterase
4-Nitrophenyl decanoate BioChemika ³99.0 % (TLC) Esterase 21497
4-Nitrophenyl hexanoate BioChemika ³98.0 % (TLC) Esterase 21610
4-Nitrophenyl octanoate BioChemika ³95.0 % (GC) Esterase 21742
4-Nitrophenyl dodecanoate BioChemika ³98.0 % (GC) Esterase 61716
4-Nitrophenyl myristate BioChemika ³95.0 % (HPLC) Esterase 70124
4-Nitrophenyl palmitate BioChemika ³98.0 % (GC) Esterase 76166
4-Nitrophenyl stearate BioChemika ³97.0 % (HPLC) Esterase 85774
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt BioChemika for fluorescence ³90% (HPCE) Esterase 74875
8-Oleoyloxypyrene-1,3,6-trisulfonic acid trisodium salt BioChemika for fluorescence Esterase 75344
L-Phenylalanine ethyl ester hydrochloride puriss. ³99.0% (AT) Esterase 78050
Resorufin acetate BioChemika for fluorescence ³98.0% (TLC) Esterase 83636
4-Methylumbelliferyl acetate Esterase probably M0883
Thymidine 3'-phosphate 4-nitrophenyl ester sodium salt BioChemika ³80.0 % (HPLC) Exonuclease 89298
N-Benzoyl-Ile-Glu-Gly-Arg p-nitroanilide hydrochloride Factor X Activated (Xa) B2291
N-Benzoyl-Ile-Glu-Gly-Arg p-nitroanilide acetate salt Factor X Activated (Xa) B7020
Boc-Ile-Glu-Gly-Arg-7-amido-4-methylcoumarin hydrochloride Factor X Activated (Xa) B9760
Factor Xa chromogenic substrate solid Factor X Activated (Xa) F3301
4-Methylumbelliferyl 4-guanidinobenzoate hydrochloride hydrate crystalline Factor X Activated (Xa) M4149
4-Methylumbelliferyl 4-guanidinobenzoate hydrochloride hydrate BioChemika for fluorescence Factor X Activated (Xa) 51010
4-Nitrophenyl 4-guanidinobenzoate hydrochloride BioChemika ³95.0 % (AT) Factor X Activated (Xa) 51015
Boc-g-benzyl-Glu-Ala-Arg-7-amido-4-methylcoumarin hydrochloride ~95% Factor Xia B4903
Boc-Phe-Ser-Arg-7-amido-4-methylcoumarin Factor XIA B6388
Pefachrome® Fxa BioChemika ³95.0 % (HPLC) Faktor Xa 77739
4-Methylumbelliferyl a-L-fucopyranoside ~98% (TLC) Fucosidase, alpha-L- M8527
4-Methylumbelliferyl a-L-fucopyranoside BioChemika for fluorescence ³98.0% (HPLC) Fucosidase, alpha-L- 69588
4-Nitrophenyl a-L-fuco­pyran­oside ³98% (TLC) Fucosidase, alpha-L- N3628
4-Nitrophenyl a-L-fuco­pyran­oside BioChemika ³98.0 % (TLC) Fucosidase, alpha-L- 73651
4-Nitrophenyl b-D-fuco­pyran­oside BioChemika ³99.0 % (HPLC) Fucosidase, alpha-L- 73652
N-Acetyl-Arg-Glu-Lys-Arg-7-amido-4-trifluoromethylcoumarin >97 % (HPLC) Furin C6483
3-Nitrotetrazolium blue chloride BioChemika ³75 % (AT) Furoyl-CoA dehydrogenase, 2- 09972
Nitrotetrazolium blue chloride BioChemika ³88 % (AT) Furoyl-CoA dehydrogenase, 2- 74030
Nitrotetrazolium blue chloride BioChemika ~10 mg/tablet nitrotetrazolium blue chloride Furoyl-CoA dehydrogenase, 2- 74031
Nitrotetrazolium blue chloride BioChemika ³98.0 % (calc. based on dry substance, AT) Furoyl-CoA dehydrogenase, 2- 74032
Tetrazolium Blue Chloride BioChemika indicator for germination, for microbiology ³90% (AT) Furoyl-CoA dehydrogenase, 2- 88190
4-Methylumbelliferyl N-acetyl-b-D-galactosaminide ³98% (TLC) Galactosaminidase, beta M9659
5-Bromo-4-chloro-3-indolyl N-acetyl-b-D-galactosaminide ~95% Galactosaminidase, N-acetyl-beta-D B3166
5-Bromo-4-chloro-3-indolyl N-acetyl-b-D-galactosaminide BioChemika ³98.0 % (TLC) Galactosaminidase, N-acetyl-beta-D 16658
4-Nitrophenyl-N-acetyl-a-D-galactosaminide BioChemika ³99.0 % (TLC) Galactosaminidase, N-acetyl-beta-D 73603
4-Nitrophenyl N-acetyl-b-D-galactosaminide BioChemika ³99.0 % (TLC) Galactosaminidase, N-acetyl-beta-D 73604
4-Nitrophenyl a-D-galacto­pyran­oside Galactosidase, alpha N0877
4-Nitrophenyl a-D-galacto­pyran­oside BioChemika ³99.0 % (HPLC) Galactosidase, alpha 73653
4-Methylumbelliferyl a-D-galactopyranoside ³98% (TLC) Galactosidase, alpha- M7633
4-Methylumbelliferyl a-D-glucopyranoside Galactosidase, alpha- M9766
4-Methylumbelliferyl a-D-galactopyranoside BioChemika for fluorescence ³96.0% (TLC) Galactosidase, alpha- 69586
4-Trifluoromethylumbelliferyl b-D-galactopyranoside BioChemika for fluorescence ³95% (HPCE) Galactosidase, beta 91879
5-Bromo-4-chloro-3-indolyl b-D-galactopyranoside BioChemika for molecular biology ³99.0% (TLC) Galactosidase, beta 16664
5-Bromo-4-chloro-3-indolyl b-D-galactopyranoside BioChemika ³98.0 % (HPLC) Galactosidase, beta 16665
5-Bromo-6-chloro-3-indolyl-b-D-galactopyranoside BioChemika ³98.0 % (HPLC) Galactosidase, beta 16669
5-Bromo-3-indolyl b-D-galactopyranoside BioChemika ³98.0 % (HPLC) Galactosidase, beta 17395
6-Bromo-2-naphthyl b-D-galactopyranoside BioChemika ³98.0 % (HPLC) Galactosidase, beta 17664
6-Chloro-3-indolyl-b-D-galactopyranoside BioChemika ³98.0 % (HPLC) Galactosidase, beta 24904
6,8-Difluoro-4-methylumbelliferyl-b-D-galactopyranoside BioChemika for fluorescence ³98.0% (TLC) Galactosidase, beta 36931
Fluorescein di(b-D-galactopyranoside) BioChemika for fluorescence ³98.0% (TLC) Galactosidase, beta 46941
Fluorescein di-(b-D-N-acetylgalactosamine) Galactosidase, beta F0802
5-Iodo-3-indolyl-b-D-galactopyranoside BioChemika ³98.0 % (HPLC) Galactosidase, beta 57897
1-Methyl-3-indolyl-b-D-galactopyranoside BioChemika ³98.0 % (HPLC) Galactosidase, beta 67610
4-Methylumbelliferyl-b-D-galactopyranoside BioChemika for fluorescence ³97.0% (HPLC) Galactosidase, beta 69590
4-Methylumbelliferyl b-D-lactopyranoside BioChemika for fluorescence ³95% (HPCE) Galactosidase, beta 69601
2-Naphthyl b-D-galactopyranoside BioChemika ³99.0 % (TLC) Galactosidase, beta 70965
4-Nitrophenyl b-D-galacto­pyran­oside ³98% (enzymatic) Galactosidase, beta N1252
2-Nitrophenyl b-D-galactopyranoside BioChemika ³99.0 % (HPLC) Galactosidase, beta 73660
4-Nitrophenyl b-D-galacto­pyran­oside BioChemika ³99.0 % (HPLC) Galactosidase, beta 73670
Phenyl-b-D-galactopyranoside BioChemika ³99.0 % (HPLC) Galactosidase, beta 78552
Resorufin b-D-galactopyranoside BioChemika for fluorescence ³95% (HPLC) Galactosidase, beta 83638
5-Bromo-4-chloro-3-indolyl b-D-galactopyranoside ³98% powder Galactosidase, beta- B4252
5-Bromo-4-chloro-3-indolyl b-D-galactopyranoside tablet Galactosidase, beta- B6024
5-Bromo-3-indolyl b-D-galactopyranoside ³98% Galactosidase, beta- B4387
2-Chloro-4-nitrophenyl-b-D-lactoside BioChemika chromophoric enzyme substrate ³90 % (HPCE) Galactosidase, beta- 25589
Fluorescein di(b-D-galactopyranoside) Galactosidase, beta- F2756
Fluorescein mono-b-D-galactopyranoside ~90% Galactosidase, beta- F6542
1-Methyl-3-indolyl-b-D-galactopyranoside ³98.0 % (HPLC) solid Galactosidase, beta- M1688
4-Methylumbelliferyl-b-D-galactopyranoside ³99% (TLC) Galactosidase, beta- M1633
Pyridoxine galactoside Galactosidase, beta- P6239
RH 421 ³95 % solid Galactosidase, beta- R8026
[2-Aminobenzoyl-Gly1]-Galanin fragment (1-10)-Lys(retro-m-Nitro-Tyr) amide, human trifluoroacetate salt >95 % (HPLC) solid Galanin degrading zinc-metallopeptidase G8916
Rac-1 protein human ³85 % recombinant, expressed in Escherichia coli buffered aqueous glycerol solution Geranylgeranyltransferase R3012
Ras, modified human ~95% (SDS-PAGE) recombinant, expressed in Escherichia coli buffered aqueous glycerol solution Geranylgeranyltransferase R0145
Laminarin from Laminaria digitata BioChemika Glucanase, beta- 61430
Cellopentaose BioChemika ³85% (HPLC) Glucanase, endo-1,4- 22163
Ostazin Brilliant Red H-3B hydroxyethyl cellulose Glucanase, endo-1,4- O6879
Hydroxyethyl-cellulose dyed with ostazin brilliant red H-3B BioChemika 10 % as dye Glucanases, endo-1,4-beta- 54295
5-Bromo-4-chloro-3-indolyl-N-acetyl-b-D-glucosaminide BioChemika ³98.0 % (TLC) Glucosaminidase, N-acetyl-beta- 16659
4-Methylumbelliferyl N-acetyl-b-D-glucosaminide dihydrate BioChemika for fluorescence ³99.0% (TLC) Glucosaminidase, N-acetyl-beta- 69585
4-Nitrophenyl N-acetyl-b-D-glucosaminide BioChemika ³99.0 % (TLC) Glucosaminidase, N-acetyl-beta- 73606
3-Nitrotetrazolium blue chloride BioChemika ³75 % (AT) Glucose Oxidase 09972
Nitrotetrazolium blue chloride BioChemika ³88 % (AT) Glucose Oxidase 74030
Nitrotetrazolium blue chloride BioChemika ~10 mg/tablet nitrotetrazolium blue chloride Glucose Oxidase 74031
Nitrotetrazolium blue chloride BioChemika ³98.0 % (calc. based on dry substance, AT) Glucose Oxidase 74032
Tetranitroblue tetrazolium chloride BioChemika ³85 % (AT) Glucose Oxidase 87961
Tetrazolium Blue Chloride BioChemika indicator for germination, for microbiology ³90% (AT) Glucose Oxidase 88190
3-Nitrotetrazolium blue chloride BioChemika ³75 % (AT) Glucose-6-phosphate dehydrogenase 09972
Nitrotetrazolium blue chloride BioChemika ³88 % (AT) Glucose-6-phosphate dehydrogenase 74030
Nitrotetrazolium blue chloride BioChemika ~10 mg/tablet nitrotetrazolium blue chloride Glucose-6-phosphate dehydrogenase 74031
Nitrotetrazolium blue chloride BioChemika ³98.0 % (calc. based on dry substance, AT) Glucose-6-phosphate dehydrogenase 74032
Tetrazolium Blue Chloride BioChemika indicator for germination, for microbiology ³90% (AT) Glucose-6-phosphate dehydrogenase 88190
4-Methylumbelliferyl a-D-glucopyranoside BioChemika for fluorescence ³99.0% (TLC) Glucosidase, alpha 69591
4-Nitrophenyl a-D-glucopyranoside ³99% Glucosidase, alpha N1377
4-Nitrophenyl a-D-glucopyranoside BioChemika ³98.0 % (HPLC) Glucosidase, alpha 73673
4-Nitrophenyl a-D-maltohexaoside BioChemika ³98.0 % (HPLC) Glucosidase, alpha 73681
4-Nitrophenyl a-D-maltopentaoside BioChemika ³98.0 % (HPLC) Glucosidase, alpha 73734
4-Nitrophenyl-6'''''-deoxy-6'''''-(2-pyridylamino)-a-D-penta-(1®4)- glucopyranoside BioChemika ³95 % (HPLC) Glucosidase, alpha 73618
4-(Trifluoromethyl)umbelliferyl-b-D-glucopyranoside BioChemika for fluorescence ³95% (HPCE) Glucosidase, beta- 91880
3-Acetylumbelliferyl b-D-glucopyranoside ³95% Glucosidase, beta- A3582
5-Bromo-4-chloro-3-indolyl b-D-glucopyranoside ~98% Glucosidase, beta- B4527
5-Bromo-4-chloro-3-indolyl b-D-glucopyranoside BioChemika ³99.0 % (TLC) Glucosidase, beta- 16667
2-Chloro-4-nitrophenyl-b-D-glucopyranoside BioChemika chromophoric enzyme substrate ³98.0 % (TLC) Glucosidase, beta- 25588
6,8-Difluoro-4-methylumbelliferyl-b-D-glucopyranoside BioChemika for fluorescence ³98.0% (TLC) Glucosidase, beta- 36937
Fluorescein di(b-D-glucuronide) Glucosidase, beta- F4551
4-Methylumbelliferyl b-D-glucopyranoside Glucosidase, beta- M3633
4-Methylumbelliferyl b-D-glucopyranoside BioChemika for fluorescence ³99.0% (HPLC) Glucosidase, beta- 69600
4-Nitrophenyl b-D-glucopyranoside Glucosidase, beta- N7006
2-Nitrophenyl b-D-glucopyranoside BioChemika ³99.0 % (HPLC) Glucosidase, beta- 73674
4-Nitrophenyl b-D-glucopyranoside BioChemika ³99.0 % (HPLC) Glucosidase, beta- 73676
Phenyl b-D-glucopyranoside BioChemika ³98.0 % (HPLC) Glucosidase, beta- 78554
Resorufin b-D-glucopyranoside BioChemika for fluorescence ³95% (HPLC) Glucosidase, beta- 83639
D-(-)-Salicin ³99% Glucosidase, beta- S0625
5-Bromo-4-chloro-3-indolyl b-D-cellobioside Glucosidases, beta- (selected) B6795
5-Bromo-4-chloro-3-indolyl--D-glucuronide Sodium salt Glucuronidase, beta-
5-Bromo-4-chloro-3-indolyl b-D-glucuronide cyclohexylammonium salt BioChemika for molecular biology ³99.0% (TLC) Glucuronidase, beta- 16666
5-Bromo-6-chloro-3-indolyl b-D-glucuronide cyclohexylammonium salt BioChemika ³98.0 % (HPLC) Glucuronidase, beta- 16671
6-Chloro-3-indolyl-b-D-glucuronide cyclohexylamine salt BioChemika ³97.0 % (HPLC) Glucuronidase, beta- 24907
6,8-Difluoro-4-methylumbelliferyl-b-D-glucuronide lithium salt BioChemika for fluorescence ³98.0% (TLC) Glucuronidase, beta- 36939
4-Methylumbelliferyl-b-D-glucuronide trihydrate BioChemika for fluorescence ³99.0% (HPLC) Glucuronidase, beta- 69602
Naphthol AS-BI b-D-glucuronide Glucuronidase, beta- N1875
Naphthol AS-BI b-D-glucuronide BioChemika for histology ³99.0% (TLC) Glucuronidase, beta- 70493
4-Nitrophenyl b-D-glucuronide BioChemika ³99.0 % (TLC) Glucuronidase, beta- 73677
Phenolphthalein b-D-glucuronide sodium salt rabbit urine Glucuronidase, beta- P0376
Phenolphthalein glucuronic acid sodium salt dihydrate rabbit urine BioChemika ³98.0 % (TLC) Glucuronidase, beta- 77637
Phenolphthalein b-D-glucuronide from rabbit urine BioChemika ³98.0 % (TLC) Glucuronidase, beta- 77636
Phenyl-b-D-glucuronide BioChemika shipped in dry ice ³99.0 % (HPLC) Glucuronidase, beta- 78555
Resorufin b-D-glucuronide sodium salt ³90% Glucuronidase, beta- R2886
5-Bromo-4-chloro-3-indolyl b-D-glucuronide cyclohexylammonium salt ³98% Glucuronidase, beta- (selection) B0522
5-Bromo-4-chloro-3-indolyl b-D-glucuronide cyclohexylammonium salt ³98% Glucuronidase, beta- (selection) B3783
5-Bromo-4-chloro-3-indolyl b-D-glucuronide cyclohexylammonium salt tablet Glucuronidase, beta- bacterial B8049
5-Bromo-4-chloro-3-indolyl b-D-glucuronide sodium salt tablet Glucuronidase, beta- bacterial B8174
5-Bromo-6-chloro-3-indolyl b-D-glucuronide cyclohexylammonium salt ³98 % Glucuronidase, beta- bacterial B4532
6-Bromo-2-naphthyl b-D-glucuronide Glucuronidase, beta- bacterial B6519
8-Hydroxyquinoline glucuronide 95-100% powder Glucuronidase, beta- bacterial H1254
4-Methylumbelliferyl b-D-glucuronide hydrate SigmaUltra Glucuronidase, beta- bacterial M5664
a-Ketoglutaric acid ³98.5% (NaOH, titration) Glutamic dehydrogenase, L- K1750
a-Ketoglutaric acid potassium salt Glutamic dehydrogenase, L- K2000
a-Ketoglutaric acid sodium salt Glutamic dehydrogenase, L- K1875
a-Ketoglutaric acid sodium salt SigmaUltra Glutamic dehydrogenase, L- K2010
Z-Leu-Leu-Glu b-naphthylamide ³98% powder Glutamylpeptidyl-peptide hydrolase C0788
L-Glutamic acid g-(3-carboxy-4-nitroanilide) ammonium salt BioChemika ³99.0 % (TLC) Glutamyltransferase, gamma- 49525
L-Glutamic acid g-(4-nitroanilide) BioChemika ³99.0 % (NT) Glutamyltransferase, gamma- 49623
Cumene hydroperoxide ~80 % Glutathione Peroxidase C0524
1-Chloro-2,4-dinitrobenzene ³98 % Glutathione S-transferase C6396
4-Chloro-7-nitrobenzofurazan BioChemika for fluorescence ³99.0% (HPLC) puriss. Glutathione S-transferase 25455
sn-Glycerol 3-phosphate bis(cyclohexylammonium) salt BioChemika ³95 % (GC) Glycerophosphate dehydrogenase, alpha- 49990
4-Nitrophenyl b-D-fuco­pyran­oside ³98% (TLC) Glycosidase, beta N3378
Z-Gly-Pro-Arg 7-amido-4-methylcoumarin hydrobromide ³85 % (TLC) powder Granzyme A C0855
Z-Gly-Pro-Arg 7-amido-4-trifluoromethylcoumarin ³97 % (HPLC) powder Granzyme A C0980
Z-Gly-Pro-Arg 4-methoxy-2-naphthylamide ³97 % (HPLC) powder Granzyme A C1105
N-Acetyl-Ile-Glu-Pro-Asp-7-amido-4-methylcoumarin ³97 % (HPLC) powder Granzyme B A6220
N-Acetyl-Ile-Glu-Pro-Asp-p-nitroaniline ³97 % (HPLC) powder Granzyme B A6470
N-Acetyl-Ile-Glu-Thr-Asp-p-nitroanilide ³95 % Granzyme B A9968
N-Acetyl-Ile-Glu-Thr-Asp-7-Amido-4-methylcoumarin ³97 % (HPLC) powder Granzyme B A4188
Boc-Ala-Ala-Asp thiobenzyl ester ³97 % (HPLC) powder Granzyme B B5305
Z-Ile-Glu-Thr-Asp 7-amido-4-trifluoromethylcoumarin ³95 % (HPLC) Granzyme B C5599
N-Acetyl-Ile-Glu-Thr-Asp-p-nitroanilide ³95 % Granzyme B A9968
Z-Ile-Glu-Thr-Asp 7-amido-4-trifluoromethylcoumarin ³95 % (HPLC) Granzyme B C5599
Guanosine 5'-triphosphate P3-[1-(2-nitrophenyl)ethyl ester] disodium salt BioChemika ³95 % (HPLC) GTPase 51125
Histamine dihydrochloride BioChemika ³99.0 % (AT) Histamine-N-methyltransferase 53300
Iodonitrotetrazolium chloride BioChemika for histol. analysis (of oxidases) ³97.0% (AT) Histidinol dehydrogenase 58030
Histone Deacetylase Substrate (Fluorogenic) >98 % (HPLC) crystalline solid Histone Deacetylase H9660
Anthranilyl-His-Lys-Ala-Arg-Val-Leu-p-nitro-Phe-Glu-Ala-Nle-Ser amide HIV Protease A0811
Anthranilyl-HIV Protease Substrate >94 % (HPLC) HIV Protease H5910
Anthranilyl-HIV Protease Substrate V >95 % (HPLC) HIV Protease H6035
Anthranilyl-HIV Protease Substrate VI >95 % (HPLC) HIV Protease H6160
HIV Protease Substrate 1 HIV Protease H6660
HIV Protease Substrate III-B, Native Sequence >98 % (HPLC) HIV Protease H5285
HIV Protease Substrate V >98 % (HPLC) HIV Protease H5410
HIV Protease Substrate VI >97 % (HPLC) HIV Protease H5535
HIV Protease Substrate VII >97 % (HPLC) HIV Protease H5660
HIV Protease Substrate VIII >96 % (HPLC) HIV Protease H5785
Boc-Val-Leu-Gly-Arg p-nitroanilide hydrobromide Horseshoe crab clotting enzyme B8391
3-(N-Ethyl-3-methylanilino)-2-hydroxypropanesulfonic acid sodium salt purum p.a. for enzymic, spectrophotometric determination of H2O2 ³98.0 % (dried material, NT) Hydrogenperoxide 04340
4-Methylumbelliferyl a-L-iduronide cyclohexylammonium salt ³99% (TLC) Iduronidase M9531
Interleukin-1b precursor fragment 110-123 ³97 % (HPLC) Interleukin-1beta converting enzyme I2775
3-Nitrotetrazolium blue chloride BioChemika ³75 % (AT) Isoquinoline 1-oxidoreductase 09972
Nitrotetrazolium blue chloride BioChemika ³88 % (AT) Isoquinoline 1-oxidoreductase 74030
Nitrotetrazolium blue chloride BioChemika ~10 mg/tablet nitrotetrazolium blue chloride Isoquinoline 1-oxidoreductase 74031
Nitrotetrazolium blue chloride BioChemika ³98.0 % (calc. based on dry substance, AT) Isoquinoline 1-oxidoreductase 74032
Tetrazolium Blue Chloride BioChemika indicator for germination, for microbiology ³90% (AT) Isoquinoline 1-oxidoreductase 88190
N-Benzoyl-Pro-Phe-Arg-p-nitroanilide hydrochloride Kallikrein B2133
Z-Phe-Arg 7-amido-4-methylcoumarin hydrochloride Kallikrein C9521
Kininogen, low molecular weight from human plasma >95% (SDS-PAGE) lyophilized powder Kallikrein K3628
Na-Benzoyl-L-arginine methyl ester bicarbonate salt Kallikrein B4256
4-Nitrophenyl 4-guanidinobenzoate hydrochloride BioChemika ³95.0 % (AT) Kallikrein 51015
Na-p-Tosyl-L-arginine methyl ester hydrochloride BioChemika for the determination of trypsin ³99.0 % (AT) Kallikrein 90170
D-Val-Leu-Arg 7-amido-4-methylcoumarin solid Kallikrein V2763
D-Val-Leu-Arg 7-amido-4-trifluoromethylcoumarin ³90 % (HPLC) solid Kallikrein V2888
a-Ketoglutaric acid ³98.5% (NaOH, titration) Ketoglutarate Dehydrogenase, alpha K1750
a-Ketoglutaric acid potassium salt Ketoglutarate Dehydrogenase, alpha K2000
a-Ketoglutaric acid sodium salt Ketoglutarate Dehydrogenase, alpha K1875
a-Ketoglutaric acid sodium salt SigmaUltra Ketoglutarate Dehydrogenase, alpha K2010
Iodonitrotetrazolium chloride BioChemika for histol. analysis (of oxidases) ³97.0% (AT) Ketolisomerase 58030
Iodonitrotetrazolium chloride BioChemika for histol. analysis (of oxidases) ³97.0% (AT) Lactate dehydrogenase 58030
4-[3-(4-Iodophenyl)-2-(2,4-dinitrophenyl)-2H-5-tetrazolio]-1,3-benzenedisulfonate sodium salt BioChemika ³90 % (HPLC) Lactic Dehydrogenase 90482
3-Nitrotetrazolium blue chloride BioChemika ³75 % (AT) Lactic Dehydrogenase 09972
Nitrotetrazolium blue chloride BioChemika ³88 % (AT) Lactic Dehydrogenase 74030
Nitrotetrazolium blue chloride BioChemika ~10 mg/tablet nitrotetrazolium blue chloride Lactic Dehydrogenase 74031
Nitrotetrazolium blue chloride BioChemika ³98.0 % (calc. based on dry substance, AT) Lactic Dehydrogenase 74032
Sodium 2-oxobutyrate BioChemika ³99.0 % (NT) Lactic Dehydrogenase 75825
a-Ketoglutaric acid sodium salt BioChemika ³98.0 % (NT) Lactic Dehydrogenase 75893
Pyruvic acid ³90 % (enzymatic) liquid Lactic Dehydrogenase P1656
Tetranitroblue tetrazolium chloride BioChemika ³85 % (AT) Lactic Dehydrogenase 87961
Tetrazolium Blue Chloride BioChemika indicator for germination, for microbiology ³90% (AT) Lactic Dehydrogenase 88190
Iodonitrotetrazolium chloride BioChemika for histol. analysis (of oxidases) ³97.0% (AT) Lactyl-CoA dehydrogenase 58030
Laminarin azure from Laminaria digitata Laminarinase L0145
L-Leucine-p-nitroanilide Leucine aminopeptidase L9125
L-Leucine 7-amido-4-methyl coumarin hydrochloride BioChemika for fluorescence Leucine aminopeptidase 61888
L-Leucine b-naphthylamide hydrochloride BioChemika substrate for the det. of leucine aminopeptidase acc. Seligmann ³97.0 % (AT) Leucine aminopeptidase 61900
N-Succinyl-Ala-Ala-Pro-Phe-7-amido-4-methylcoumarin Leucine Aminopeptidase S9761
N-Succinyl-Phe-Gly-Leu-b-naphthylamide LH-RH-degrading endopeptidase S6768
Boc-Ser(Bzl)-Gly-Arg-NH-Np.HCl BioChemika ³98.0 % (HPLC) Limulus Clotting Enzyme 76556
1,2-Di-O-decanoyl-rac-glycero-3-glutaric acid-resorufin labeled Lipase D7414
Fluorescein dibutyrate BioChemika for fluorescence ³98.0% (HPCE) Lipase 46942
Fluorescein dilaurate BioChemika for fluorescence ³97.0% (HPCE) Lipase 46943
Indoxyl acetate BioChemika ³97.0% (TLC) Lipase 57420
Lipase Substrate BioChemika for the titrimetric determination of enzyme activity Lipase 62314
4-Methylumbelliferyl butyrate BioChemika for fluorescence ³95% (HPCE) Lipase 19362
4-Methylumbelliferyl heptanoate BioChemika for fluorescence ³98.0% (TLC) Lipase 75230
4-Methylumbelliferyl oleate BioChemika for fluorescence ³90% (HPCE) Lipase 75164
4-Methylumbelliferyl palmitate BioChemika for fluorescence ³98.0% (TLC) Lipase 76163
2-Naphthyl laurate BioChemika substrate for the det. of lipase ³99.0 % (GC) Lipase 61710
Resorufin butyrate BioChemika for fluorescence ³95% (TLC) Lipase 83637
4-Trifluoromethylumbelliferyl oleate Lipase T5069
Alizarin-3-methyliminodiacetic acid Lipid Peroxidation A3882
Firefly tails (lanterns), desiccated Luciferase FFT
Fireflies, whole, desiccated Luciferase FFW
Firefly Lantern Extract powder Luciferase F3641
Firefly Lantern Extract Luciferase F8021
FLE50 FIREFLY LANTERN EXTRACT Luciferase FLE-50
Firefly diluent lyophilized powder Luciferase F3766
D-Luciferin BioChemika ³96.0 % (HPLC) Luciferase 62643
Luciferin 6'-ethyl ether sodium salt Luciferase L9007
Luciferin 6'-methyl ether sodium salt Luciferase L1147
D-Luciferin synthetic SigmaUltra ³99% (HPLC) Luciferase L6152
D-Luciferin sodium salt lyophilized powder synthetic Luciferase L6882
D-Luciferin sodium salt BioChemika ³98.0 % (HPLC) Luciferase 62645
D-Luciferin synthetic Luciferase L9504
D-Luciferin potassium salt Luciferase L7538
Peptidoglycan dyed with Remazol Brilliant Blue R Staphylococcus aureus BioChemika Lysostaphin 77145
Peptidoglycan from Micrococcus luteus BioChemika Lysostaphin 53243
4-Methylumbelliferyl b-D-N,N',N''-triacetylchitotrioside Lysozyme M5639
4-Methylumbelliferyl b-D-N,N',N''-triacetylchitotrioside BioChemika for fluorescence 90% Lysozyme 69615
4-Nitrophenyl b-D-N,N',N''-triacetylchitotriose Lysozyme N8638
Peptidoglycan dyed with Remazol Brilliant Blue R Staphylococcus aureus BioChemika Lysozyme 77145
Peptidoglycan from Micrococcus luteus BioChemika Lysozyme 53243
Yeast BioChemika powder color slightly brown Lyticase 51475
Oxaloacetic acid BioChemika ³98.0 % (T) Malate dehydrogenase 75660
4-Methylumbelliferyl a-D-mannopyranoside ~98% (TLC) Mannosidase, alpha- M3657
4-Methylumbelliferyl a-D-mannopyranoside BioChemika for fluorescence ³99.0% (TLC) Mannosidase, alpha- 69604
4-Nitrophenyl a-D-mannopyranoside BioChemika ³99.0 % (TLC) Mannosidase, alpha- 73726
4-Nitrophenyl a-D-mannopyranoside Mannosidase, alpha- N2127
4-Nitrophenyl b-D-mannopyranoside ³98% Mannosidase, Beta- N1268
4-Nitrophenyl b-D-mannopyranoside BioChemika ³99.0 % (TLC) Mannosidase, Beta- 73727
N-(2,4-Dinitrophenyl)-Pro-Leu-Gly-Met-Trp-Ser-Arg solid Matrix metalloproteinase D4315
7-Methoxycoumarin-4-acetyl-Pro-Leu-Gly-Leu-b-(2,4-dinitrophenylamino)Ala-Ala-Arg amide ³97% (HPLC) Matrix metalloproteinase M6412
7-Methoxycoumarin-4-acetyl-Pro-Leu-Ala-Nva-L-(2,4-dinitrophenyl)diaminopropio nyl-Ala-Arg-amide solid Matrix metalloproteinase-3 M3439
N-(2,4-Dinitrophenyl)-Pro-Leu-Gly-Leu-Trp-Ala-D-Arg amide Matrix metalloproteinases MMP-1 and MMP-9 D2293
N-(2,4-Dinitrophenyl)-Pro-Leu-Gly-Leu-Trp-Ala-D-Arg amide Matrix metalloproteinases MMP-1 and MMP-9 D2293
DABCYL-[Nle1077]-Collagen Type III a 1 Chain fragment 1070-1077–EDANS, Human >96 % (HPLC) Type III a 1 chain Metalloproteinase C9732
7-Methoxycoumarin-4-acetyl-Arg-Pro-Leu-Ala-Leu-Trp-Arg-L-a,b-diaminopropionyl(2,4-dinitrophenyl) amide >97 % (HPLC) solid Metalloproteinase M0813
N-Acetyl-S-geranylgeranyl-L-cysteine ³95% ethanol solution Methyltransferase, S-Adenosylmethionine-dependent A6331
Kynuramine dihydrobromide crystalline Monoamine Oxidase K3250
Tryptamine hydrochloride BioChemika ³98.0 % (AT) Monoamine oxidase 93650
Tyramine BioChemika for fluorescence ³99.0% (TLC) Monoamine oxidase 93810
7-Ethoxycoumarin BioChemika for fluorescence ³99.0% (HPLC) Monooxygenase 02644
1-Carbamoylmethyl-3-carbamoyl-1,4-dihydropyrimidine NAD(P)H quinone oxidoreductase 2 C1485
CB 1954 powder NAD(P)H quinone oxidoreductase 2 C2235
1-(3-Sulfonatopropyl)-3-carbamoyl-1,4-dihydropyrimidine ³85 % (TLC) solid NAD(P)H quinone oxidoreductase 2 S0190
1-Naphthyl acetate BioChemika ³99.0 % (HPLC) Naphthyl Acetate Esterase 46010
Primisulfuron-methyl PESTANAL®, analytical standard Naphthyl Acetate Esterase 46010
2-Naphthyl acetate BioChemika substrate for the det. of esterase ³99.0 % (GC) Naphthyl Acetate Esterase 46020
5-Bromo-4-chloro-3-indolyl a-D-N-acetylneuraminic acid sodium salt ³90% Neuraminidase B4666
2'-(4-Methylumbelliferyl)-a-D-N-acetylneuraminic acid sodium salt hydrate ³95% (HPLC) Neuraminidase M8639
2'-(4-Methylumbelliferyl)-a-D-N-acetylneuraminic acid sodium salt hydrate BioChemika for fluorescence ³97.0% (HPLC) Neuraminidase 69587
2-O-(p-Nitrophenyl)-a-D-N-acetylneuraminic acid ³95% Neuraminidase N1516
L-Arginine reagent grade ³98% (TLC) powder Nitric Oxide Synthase A5006
L-Arginine L-glutamate salt Nitric Oxide Synthase A5756
L-Arginine monohydrochloride reagent grade ³98% (TLC) powder Nitric Oxide Synthase A5131
NITRIC OXIDE SYNTHASE DETECTION SYSTEM, FLUOROMETRIC, FOR CELL-ASSOCIATED NOS NITRIC OXIDE SYNTHASE FCANOS-1
NOC-5 ³95% Nitric Oxide Synthase/NO Donor A5456
3,3-Bis(aminoethyl)-1-hydroxy-2-oxo-1-triazene ³95% Nitric Oxide Synthase/NO Donor A5581
Diethylamine NONOate diethylammonium salt Nitric Oxide Synthase/NO Donor D5431
Diethylamine NONOate sodium salt crystalline Nitric Oxide Synthase/NO Donor D184
Dipropylenetriamine NONOate Nitric Oxide Synthase/NO Donor D5057
(±)-(E)-4-Ethyl-2-[(E)-hydroxyimino]-5-nitro-3-hexenamide ³98 % Nitric Oxide Synthase/NO Donor E2895
(±)-(E)-4-Ethyl-2-[(Z)-hydroxyimino]-5-nitro-3-hexen-1-yl-nicotinamide ³97 % Nitric Oxide Synthase/NO Donor E3020
(±)-(E)-2-[(E)-Hydroxyimino]-6-methoxy-4-methyl-5-nitro-3-hexenamide ³95 % (HPLC) Nitric Oxide Synthase/NO Donor H9528
1-Hydroxy-3-methyl-3-(methylaminopropyl)-2-oxo-1-triazene ³90% (HPLC) Nitric Oxide Synthase/NO Donor H9778
Isosorbide dinitrate powder Nitric Oxide Synthase/NO Donor I1759
MAHMA NONOate Nitric Oxide Synthase/NO Donor M1555
Molsidomine Nitric Oxide Synthase/NO Donor M2901
3-Morpholinosydnonimine hydrochloride Nitric Oxide Synthase/NO Donor M5793
3-Morpholinosydnonimine hydrochloride solid Nitric Oxide Synthase/NO Donor M184
PAPA NONOate Nitric Oxide Synthase/NO Donor P8102
4-Phenyl-3-furoxancarbonitrile Nitric Oxide Synthase/NO Donor P1726
Proli NONOate Nitric Oxide Synthase/NO Donor P7977
S150 SPERMINE/NITRIC OXIDE COMPLEX Nitric Oxide Synthase/NO Donor S150
SE 175 ³97 % Nitric Oxide Synthase/NO Donor S2189
S-Nitroso-N-acetylpenicillamine ³97% Nitric Oxide Synthase/NO Donor N3398
S-Nitrosoglutathione ³97% Nitric Oxide Synthase/NO Donor N4148
Sodium nitroprusside dihydrate Nitric Oxide Synthase/NO Donor S0501
Streptozocin ³75% as a-anomer ³98% (HPLC) powder Nitric Oxide Synthase/NO Donor S0130
Sulfo NONOate disodium salt Nitric Oxide Synthase/NO Donor S8432
Lithium carbamoylphosphate dibasic BioChemika ~85 % (enzymatic) Ornithine Carbamoyl-transferase 21780
Oxaloacetic acid BioChemika ³98.0 % (T) Oxaloacetate decarboxylase 75660
Iodonitrotetrazolium chloride BioChemika for histol. analysis (of oxidases) ³97.0% (AT) Oxidase 58030
S-Benzyl-L-cysteine 4-nitroanilide BioChemika ³98.0 % (NT) Oxytocinase 13340
L-Cystine-di-2-naphthylamide BioChemika ³98.0 % (HPLC) Oxytocinase 30260
P to Z Back to top
Substrate Enzyme Product # 
PRAK Substrate Peptide 1 mg/mL in water ³98 % (HPLC) aqueous solution P38 Regulated/Activated Protein Kinase P0240
PAK I PEPTIDE SUBSTRATE PAK, gamma- (PAK I) A-284
Z-L-arginine 7-amido-4-methylcoumarin hydrochloride BioChemika for fluorescence ³90% (TLC) Papain 95938
Na-Benzoyl-L-arginine 4-nitroanilide hydrochloride BioChemika ³98.0 % (enzymatic) Papain 12915
Na-Benzoyl-L-arginine-7-amido-4-methylcoumarin hydrochloride Papain B7260
Fibrin Blue suitable for substrate for pepsin at low pH (High blanks result at high pH) Pepsin F5255
Z-Glu-Tyr BioChemika substrate for the det. of pepsin ³99.0 % (HPLC) Pepsin 96150
Phe-Ala-Ala-Phe(4-NO2)-Phe-Val-Leu (4-pyridylmethyl) ester BioChemika ³98.0 % (TLC) Pepsin 77431
N-Benzoyl-L-tyrosyl-4-aminobenzoic acid Sodium salt Peptidase 13136
N-[3-(2-Furyl)acryloyl]-Phe-Gly-Gly BioChemika ³98.0 % (TLC) Peptidase 48176
Naphthol AS-D BioChemika ³99.0 % (HPLC) Peptidase 70489
L-Pyroglutamic acid 2-naphthylamide BioChemika ³99.0 % (TLC) Peptidase 83176
N-Succinyl-Ala-Ala-Pro-Phe p-nitroanilide Peptidyl prolyl isomerase S7388
N-Oleoylglycine ³98 % Peptidylglycine a-amidating enzyme. O9762
N-(4-Aminobutyl)-N-ethylisoluminol ³90% Peroxidase A0156
N-(6-Aminohexyl)-N-ethylisoluminol Peroxidase A1661
3-(4-Hydroxyphenyl)propionic acid Peroxidase H6386
2,2'-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt BioChemika 10 mg/tablet ABTS Peroxidase 11565
Luminol sodium salt Peroxidase A4685
Luminol ³97% (HPLC) Peroxidase A8511
3-Amino-9-ethylcarbazole ~90% Peroxidase A5754
3-Amino-9-ethylcarbazole tablet Peroxidase A6926
4-Aminophthalhydrazide monohydrate Peroxidase A8264
3-Amino-9-ethylcarbazole BioChemika 20 mg/tablet AEC Peroxidase 03005
4-Aminoantipyrine reagent grade Peroxidase A4382
5-Aminosalicylic acid BioChemika ~100 mg/tablet 5-aminosalicylic acid (enzym.) tablets Peroxidase 09407
5-Aminosalicylic acid tablet Peroxidase A6178
5-Aminosalicylic acid ~99% Peroxidase A3537
2,2'-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) Liquid Substrate System Peroxidase A3219
2,2'-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt ~98% (TLC) Peroxidase A1888
2,2'-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt tablet 10 mg substrate per tablet Peroxidase A9941
CHEMILUMINESCENT PEROXIDASE SUBSTRATE Peroxidase CPS-1
4-Chloro-1-naphthol tablet Peroxidase C6788
2-Chloro-5,5-dimethyl-1,3-cyclohexanedione BioChemika suitable as substrate for chloroperoxidase ³98.0 % (AT) Peroxidase 24368
4-Chloro-1-naphthol BioChemika for analytical purposes ³99.0% (HPLC) Peroxidase 25328
4-Chloro-1-naphthol BioChemika ³95.0% (HPLC) tablets (~32 mg each) purum Peroxidase 25329
4-Chloro-1-naphthol purum ³98.0% (T) Peroxidase 25330
4-Chloro-7-nitrobenzofurazan BioChemika for fluorescence ³99.0% (HPLC) puriss. Peroxidase 25455
3,3'-Diaminobenzidine (DAB) Enhanced Liquid Substrate System tetrahydrochloride Peroxidase D3939
3,3'-Diaminobenzidine (DAB) Enhanced Liquid Substrate System tetrahydrochloride for Membrane Applications Peroxidase D6815
3,3'-Diaminobenzidine tetrahydrochloride tablet 10 mg substrate per tablet Peroxidase D5905
o-Dianisidine dihydrochloride tablet 10 mg substrate per tablet Peroxidase D9154
o-Dianisidine Peroxidase D9143
o-Dianisidine dihydrochloride crystalline Peroxidase D3252
Dicarboxidine dihydrochloride Peroxidase D5907
Sodium 3,5-dichloro-2-hydroxybenzenesulfonate BioChemika ³98.0 % (TLC) Peroxidase 35867
Dihydrorhodamine 123 BioChemika for fluorescence ³95% (HPLC) Peroxidase 37370
o-Dianisidine dihydrochloride BioChemika ~10 mg/tablet o-dianisidine dihydrochloride Peroxidase 33435
3-(Dimethylamino)benzoic acid ³97% (HPLC) Peroxidase D0787
Guaiacol Peroxidase G5502
Iodonitrotetrazolium chloride BioChemika for histol. analysis (of oxidases) ³97.0% (AT) Peroxidase 58030
Neotetrazolium chloride form BioChemika for microbiology for the detection of reducing fermentation systems ³75% (T) Peroxidase 72150
3-Nitrotetrazolium blue chloride BioChemika ³75 % (AT) Peroxidase 09972
Nitrotetrazolium blue chloride BioChemika ³88 % (AT) Peroxidase 74030
Nitrotetrazolium blue chloride BioChemika ~10 mg/tablet nitrotetrazolium blue chloride Peroxidase 74031
Nitrotetrazolium blue chloride BioChemika ³98.0 % (calc. based on dry substance, AT) Peroxidase 74032
o-Phenylenediamine BioChemika for fluorescence free base Peroxidase 78411
o-Phenylenediamine dihydrochloride BioChemika for fluorescence Peroxidase 78441
o-Phenylenediamine dihydrochloride BioChemika Peroxidase 78443
o-Phenylenediamine dihydrochloride BioChemika Peroxidase 78446
o-Phenylenediamine dihydrochloride BioChemika Peroxidase 78449
o-Phenylenediamine dihydrochloride crystalline Peroxidase P1526
o-Phenylenediamine dihydrochloride tablet 60 mg substrate per tablet Peroxidase P1063
o-Phenylenediamine dihydrochloride tablet 15 mg substrate per tablet Peroxidase P4664
o-Phenylenediamine dihydrochloride tablet 1 mg substrate per tablet Peroxidase P6662
o-Phenylenediamine dihydrochloride tablet 2 mg substrate per tablet Peroxidase P6787
o-Phenylenediamine dihydrochloride tablet 5 mg substrate per tablet Peroxidase P6912
o-Phenylenediamine dihydrochloride tablet 30 mg substrate per tablet Peroxidase P8412
o-Phenylenediamine dihydrochloride tablet 4 mg substrate per tablet Peroxidase P8787
SIGMA FASTä OPD tablet Peroxidase P9187
o-Phenylenediamine solid Peroxidase P9029
o-Phenylenediamine tablet 20 mg substrate per tablet Peroxidase P5412
o-Phenylenediamine dihydrochloride tablet 20 mg substrate per tablet Peroxidase P7288
o-Phenylenediamine dihydrochloride tablet 3 mg substrate per tablet Peroxidase P8806
o-Phenylenediamine dihydrochloride tablet 5 mg substrate per tablet Peroxidase P3804
o-Phenylenediamine dihydrochloride tablet 10 mg substrate per tablet Peroxidase P8287
Pyrogallol Peroxidase P0381
Pyrogallol ACS reagent Peroxidase P2923
SIGMAFAST™ 3,3'-Diaminobenzidine tablets tablet To prepare 1 mL Peroxidase D4168
SIGMAFAST™ 3,3'-Diaminobenzidine tablets tablet To prepare 5 mL Peroxidase D4293
SIGMAFAST™ 3,3'-Diaminobenzidine tablets tablet To prepare 15 mL Peroxidase D4418
SIGMAFAST™ DAB with Metal Enhancer tablet Peroxidase D0426
3,3',5,5'-Tetramethylbenzidine dihydrochloride Peroxidase T8768
3,3',5,5'-Tetramethylbenzidine (TMB) Liquid Substrate System for ELISA Peroxidase T0440
3,3',5,5'-Tetramethylbenzidine (TMB) Liquid Substrate System for Membranes Peroxidase T0565
3,3',5,5'-Tetramethylbenzidine Liquid Substrate, Slow Kinetic Form, for ELISA Peroxidase T4319
3, 3',5 ,5'-Tetramethylbenzidine Liquid Substrate, Supersensitive, for ELISA Peroxidase T4444
3,3',5,5'-Tetramethylbenzidine ³98% (TLC) Peroxidase T2885
3,3',5,5'-Tetramethylbenzidine (TMB) Liquid Substrate System Peroxidase T8665
3,3',5,5'-Tetramethylbenzidine red liquid substrate system liquid Peroxidase T1693
3,3',5,5'-Tetramethylbenzidine dihydrochloride tablet 1 mg substrate per tablet Peroxidase T3405
3,3',5,5'-Tetramethylbenzidine tablet 1 mg substrate per tablet Peroxidase T5525
3,3',5,5'-Tetramethylbenzidine Liquid Substrate, Super Slow, for ELISA Peroxidase T5569
Tetranitroblue tetrazolium chloride BioChemika ³85 % (AT) Peroxidase 87961
Tetrazolium Blue Chloride BioChemika indicator for germination, for microbiology ³90% (AT) Peroxidase 88190
Tetrazolium Violet BioChemika ³94 % (AT) Peroxidase 88193
Thiazolyl blue formazan BioChemika Peroxidase 88417
Thiazolyl Blue Tetrazolium Bromide BioChemika ³97.0 % (enzymatic) Peroxidase 88415
trans-5-Phenyl-4-pentenyl hydroperoxide ³90 % Solution in ethanol Peroxidase P6238
2,3,5-Triphenyltetrazolium chloride BioChemika ³99.0 % (AT) Peroxidase 93140
Hydrogen peroxide–Urea adduct ³97% Peroxidase U1753
Hydrogen peroxide–Urea adduct tablet Peroxidase U8879
Umbelliferyl Arachidonate ethanol solution Phopholipase A2 U0383
O-(4-Nitrophenylphosphoryl)choline Phopholipase C N5879
1-Naphthyl 4-phenylazophenyl phosphate purum ³98.0% (T) Phosphodiesterase 10922
2-Naphthyl 4-phenylazophenyl phosphate BioChemika ³98.0 % (T) Phosphodiesterase 10923
Thymidine 3'-phosphate 4-nitrophenyl ester sodium salt BioChemika ³80.0 % (HPLC) Phosphodiesterase II 89298
4-Nitrophenyl phenylphosphonate Phosphodiesterase, 5'-Nucleotide N2881
Xanthosine 3',5'-cyclic monophosphate ³98 % (HPLC) solid Phosphodiesterase, guanosine-3',5'-monophosphate X3003
Phospho(enol)pyruvic acid monopotassium salt BioChemika ³98.0 % (PEP-K, enzymatic) Phosphoenolpyruvate carboxylase 79415
Phospho(enol)pyruvic acid monosodium salt monohydrate BioChemika ³98.0 % (NT) Phosphoenolpyruvate carboxylase 79418
Phospho(enol)pyruvic acid tri(cyclohexylammonium) salt BioChemika crystallized ³98.0 % Phosphoenolpyruvate carboxylase 79430
Phospho(enol)pyruvic acid trisodium salt heptahydrate BioChemika ³98.0 % (enzymatic) Phosphoenolpyruvate carboxylase 79435
6-Phosphogluconic acid barium salt hydrate BioChemika ³95.0 % (TLC) Phosphogluconate Dehydrogenase 79450
6-Phosphogluconic acid tri(cyclohexylammonium) salt BioChemika ³99.0 % (TLC) Phosphogluconate Dehydrogenase 79458
6-Phosphogluconic acid trisodium salt BioChemika ³97.0 % (calc. based on dry substance, enzymatic) Phosphogluconate Dehydrogenase 79455
O-(4-Nitrophenylphosphoryl)choline BioChemika ³98.0 % (TLC) Phospholipase C 73739
1,2-Dipalmitoylphosphatidylinositol 4,5-diphosphate ³98 % solid Phospholipase C PI-specific P7115
1,2-Dipalmitoylphosphatidylinositol 3,4-diphosphate ³98 % solid Phospholipase C PI-specific P6990
Sodium phenyl phosphate dibasic dihydrate BioChemika ³95.0 % (NT) Phosphotransferase 71900
D-Ala-Leu-Lys-7-amido-4-methylcoumarin 95% (HPLC) Plasmin A8171
Boc-Val-Leu-Lys-7-amido-4-methylcoumarin Plasmin B1136
Z-Ala-Ala-Lys 4-methoxy-b-naphthylamide formate salt Plasmin C2147
Gly-Arg-p-nitroanilide dihydrochloride Plasmin G8148
D-Ile-Phe-Lys p-nitroanilide ³95 % (HPLC) Plasmin I6886
4-Nitrophenyl 4-guanidinobenzoate hydrochloride BioChemika ³95.0 % (AT) Plasmin 51015
N-(p-Tosyl)-Gly-Pro-Lys 4-nitroanilide acetate salt Plasmin T6140
Na-p-Tosyl-L-arginine methyl ester hydrochloride BioChemika for the determination of trypsin ³99.0 % (AT) Plasmin 90170
N-(p-Tosyl)-Gly-Pro-Lys 4-nitroanilide acetate salt BioChemika ³99.0 % (TLC) Plasmin 90178
D-Val-Leu-Lys 4-nitroanilide dihydrochloride Plasmin V0882
D-Val-Leu-Lys 4-nitroanilide dihydrochloride BioChemika ³98.0 % (HPLC) Plasmin 94680
Porphobilinogen BioChemika (synth.) ³97.0 % (HPLC) Porphobilinogen deaminase 81480
Arg-Glu(EDANS)-[Asn670, Leu671]-Amyloid b/A4 Precursor Protein 770 Fragment 669-674-(DABCYL)Lys-Arg ³96 % (HPLC) solid pro-beta-secretase (pro-memapsin 2) A2847
Mca-[Asn670, Leu671]-Amyloid b/A4 Precursor Protein 770 fragment 667-675-(Dnp)Lys amide >96 % (HPLC) solid pro-beta-secretase (pro-memapsin 2) M1063
L-Proline-2-naphthylamide hydrochloride BioChemika ³99.0 % (TLC) Proline aminopeptidase 81742
L-Proline-2-naphthylamide hydrochloride BioChemika ³99.0 % (TLC) Proline beta-naphthylamidase 81742
2-Aminobenzoyl-Gly-Ala-Ala-Pro-Phe-Tyr(NO2)-Asp trifluoroacetate salt ³90 % (HPLC) Proline SpecificEndopeptidase A0222
Z-Gly-Pro 7-amido-4-methylcoumarin BioChemika for fluorescence ³97.0% (CHN) Prolyl endopeptidase 96280
Z-Gly-Pro-2-naphthylamide BioChemika for fluorescence ³97.0% (TLC) Prolyl endopeptidase 96283
Z-Gly-Pro-4-nitroanilide BioChemika ³99.0 % (TLC) Prolyl endopeptidase 96286
1-Naphthyl phosphate disodium salt BioChemika ³99.0 % (HPLC) Prostatic acid phosphatase 71105
N-(3-[2-Furyl]acryloyl)-Ala-Phe amide Protease F2380
N-(3-[2-Furyl]acryloyl)-Gly-Leu amide Protease F7383
Albumin, Fluorescein isothiocyanate Conjugate bovine Protease A9771
Anthranilyl-Asp-Phe-Arg-Leu-Phe-Ala-Phe-nitro-Tyr-Asp ³97% (HPLC) Protease A0324
Azo dye-impregnated collagen suitable for non-specific protease substraterate of dye release may vary from lot to lot (Dye-impregnated hide powder.) Protease A4341
Azoalbumin Protease A2382
Azocasein Protease A2765
Azo-Casein BioChemika Protease 11610
Azo-Casein sodium salt BioChemika lyophilized Protease 11615
Boric anhydride granulated Protease 11615
Casein, Resorufin-labeled BioChemika Protease 22097
Z-Gly-Gly-Leu p-nitroanilide Protease C3022
Fibrin Blue suitable for substrate for pepsin at low pH (High blanks result at high pH) Protease F5255
L-Leucine-7-amido-4-methylcoumarin hydrochloride Protease L2145
N,N-Dimethylcasein BioChemika for spectrophotometric det. of enzyme activity Protease 39890
N-Succinyl-Gly-Gly-Phe-p-nitroanilide Protease B S1899
Anthranilyl-Ala-Phe-Ala-Phe-Glu-Val-Phe-nitro-Tyr-Asp ³90% (HPLC) Proteases glutamate-specific A3567
Z-Leu-Leu-Glu-7-amido-4-methylcoumarin ³95 % solid Proteasome C0483
Z-Leu-Leu-Leu-7-amido-4-methylcoumarin ³95 % (TLC and HPLC) solid Proteasome C0608
N-Succinyl-Leu-Leu-Val-Tyr-7-amido-4-trifluoromethylcoumarin ³98 % (TLC) solid Proteasome S4939
Boc-Leu-Ser-Thr-Arg-7-amido-4-methylcoumarin Protein C. B4636
[Trp4]-Kemptide ³97% (HPLC) Protein Kinase A L3523
[Val6, Ala7]-Kemptide ³97% (HPLC) Protein Kinase A K2877
Arg-Thr-Lys-Arg-Ser-Gly-Ser-Val-Tyr-Glu-Pro-Leu-Lys-Ile ³97 % (HPLC) powder Protein Kinase A A3317
Kemptide acetate salt ³97% (HPLC) powder Protein Kinase A K1127
BAD from mouse ³70 % (SDS-PAGE) recombinant, expressed in Escherichia coli buffered aqueous glycerol solution Protein Kinase B B1682
Myelin Basic Protein Fragment 4-14 ³95% (HPLC) Protein Kinase C M6913
[Ser25]-Protein Kinase C fragment 19-31 ³97 % (HPLC) Protein Kinase C P1835
EGF Receptor Fragment 651-658 >95 % (HPLC) Protein Kinase C E5527
Lys-Arg-Thr-Leu-Arg-Arg trifluoroacetate salt ³97 % (HPLC) Protein Kinase C L9905
Myelin Basic Protein Fragment 104-118 synthetic lyophilized powder Protein Kinase C M8184
Neurogranin Fragment 28-43 ³97 % (HPLC) Protein Kinase C N7279
[pGlu4]-Myelin basic protein fragment 4-14 ³97 % (HPLC) Protein Kinase C P2186
Pro-Leu-Ser-Arg-Thr-Leu-Ser-Val-Ala-Ala-Lys-Lys ³97% (HPLC) Protein Kinase C P5307
Val-Arg-Lys-Arg-Thr-Leu-Arg-Arg-Leu ³97 % (HPLC) Protein Kinase C V2131
Arg-Lys-Arg-Ser-Arg-Ala-Glu lyophilized powder Protein kinase G A6833
Arg-Lys-Arg-Ser-Arg-Lys-Glu lyophilized powder Protein kinase G A8457
Protein Tyrosine Phosphatase Substrate Monophosphate ³97% (HPLC) Protein Tyrosine Phosphatase P6462
L-a-Phosphatidyl-D-myo-inositol 3,4,5-triphosphate, dioctanoyl PTEN P9953
L-Pyroglutamic acid 7-amido-4-methylcoumarin BioChemika ³99.0 % (HPLC) Pyroglutamyl aminopeptidase 83174
L-Pyroglutamic acid 2-naphthylamide BioChemika ³99.0 % (TLC) Pyroglutamyl Peptidase 83176
Esculin sesquihydrate BioChemika ³98.0 % (HPLC) Pyrrolidonyl peptidase 02350
Phospho(enol)pyruvic acid monopotassium salt BioChemika ³98.0 % (PEP-K, enzymatic) Pyruvate kinase 79415
Phospho(enol)pyruvic acid monosodium salt monohydrate BioChemika ³98.0 % (NT) Pyruvate kinase 79418
Phospho(enol)pyruvic acid tri(cyclohexylammonium) salt BioChemika crystallized ³98.0 % Pyruvate kinase 79430
Phospho(enol)pyruvic acid trisodium salt heptahydrate BioChemika ³98.0 % (enzymatic) Pyruvate kinase 79435
3-Nitrotetrazolium blue chloride BioChemika ³75 % (AT) Quinaldate 4-oxidoreductase 09972
Nitrotetrazolium blue chloride BioChemika ³88 % (AT) Quinaldate 4-oxidoreductase 74030
Nitrotetrazolium blue chloride BioChemika ~10 mg/tablet nitrotetrazolium blue chloride Quinaldate 4-oxidoreductase 74031
Nitrotetrazolium blue chloride BioChemika ³98.0 % (calc. based on dry substance, AT) Quinaldate 4-oxidoreductase 74032
Tetrazolium Blue Chloride BioChemika indicator for germination, for microbiology ³90% (AT) Quinaldate 4-oxidoreductase 88190
3-Fluoro-3-deoxy-D-glucose BioChemika ³99.0 % (TLC) Reductase 46865
Renin Substrate 1 Renin R8276
N-Benzoyl-Phe-Val-Arg-p-nitroanilide hydrochloride BioChemika ³99.0 % (TLC) Reptilase 13042
Ribonucleic acid from baker's yeast (S. cerevisiae) BioChemika ~18 A260/mg RNase 83847
NFAT 1 from mouse ³90% (SDS-PAGE) recombinant, expressed in Escherichia coli N-terminal 6-His tagged Truncated sequence amino acids 1-297 buffered aqueous glycerol solution SAPK2a/p38a N9659
TNPAL-Sphingomyelin 1 mg/mL in chloroform: methanol (2:1 v/v) Sphingomyelinase T1014
Na-p-Tosyl-L-arginine methyl ester hydrochloride BioChemika for the determination of trypsin ³99.0 % (AT) Subtilisin 90170
2-(2'-Benzothiazolyl)-5-styryl-3-(4'-phthalhydrazidyl)tetrazolium chloride 90-95% Succinic dehydrogenase B8131
Iodonitrotetrazolium chloride BioChemika for histol. analysis (of oxidases) ³97.0% (AT) Succinic Dehydrogenase 58030
4-[3-(4-Iodophenyl)-2-(2,4-dinitrophenyl)-2H-5-tetrazolio]-1,3-benzenedisulfonate sodium salt BioChemika ³90 % (HPLC) Succinic Dehydrogenase 90482
3-Nitrotetrazolium blue chloride BioChemika ³75 % (AT) Succinic Dehydrogenase 09972
Nitrotetrazolium blue chloride BioChemika ³88 % (AT) Succinic Dehydrogenase 74030
Nitrotetrazolium blue chloride BioChemika ~10 mg/tablet nitrotetrazolium blue chloride Succinic Dehydrogenase 74031
Nitrotetrazolium blue chloride BioChemika ³98.0 % (calc. based on dry substance, AT) Succinic Dehydrogenase 74032
Tetranitroblue tetrazolium chloride BioChemika ³85 % (AT) Succinic Dehydrogenase 87961
Tetrazolium Blue Chloride BioChemika indicator for germination, for microbiology ³90% (AT) Succinic Dehydrogenase 88190
2,3,5-Triphenyltetrazolium chloride BioChemika ³99.0 % (AT) Succinic Dehydrogenase 93140
1,3,5-Triphenyltetrazolium formazan BioChemika ³97.0 % (UV) Succinic Dehydrogenase 93145
5-Bromo-4-chloro-3-indolyl sulfate potassium salt BioChemika ³99.0 % (T) Sulfatase 16673
5-Bromo-6-chloro-3-indolyl sulfate potassium salt BioChemika ³99.0 % (HPLC) Sulfatase 16674
4-Nitrocatechol sulfate dipotassium salt monohydrate BioChemika ³97.0 % (T) Sulfatase 56020
4-Methylumbelliferyl sulfate potassium salt BioChemika for fluorescence ³99.0% (HPLC) Sulfatase 69610
Phenolphthalein disulfate tripotassium salt BioChemika ³99.0 % (TLC) Sulfatase 77635
3-Nitrotetrazolium blue chloride BioChemika ³75 % (AT) Superoxid dismutase 09972
Nitrotetrazolium blue chloride BioChemika ³88 % (AT) Superoxid dismutase 74030
Nitrotetrazolium blue chloride BioChemika ~10 mg/tablet nitrotetrazolium blue chloride Superoxid dismutase 74031
Nitrotetrazolium blue chloride BioChemika ³98.0 % (calc. based on dry substance, AT) Superoxid dismutase 74032
Tetrazolium Blue Chloride BioChemika indicator for germination, for microbiology ³90% (AT) Superoxid dismutase 88190
Methyl gallate purum antioxidant ³98.0% (GC) Tannase 48690
7-Methoxycoumarin-4-acetyl-[Asn670, Leu671]-Amyloid b/A4 Precursor Protein 770 Fragment 667-676-(2,4-dinitrophenyl)Lys-Arg-Arg amide ³90 % (HPLC) Thimet oligopeptidase (TOP) A1472
7-Methoxycoumarin-4-acetyl-[Asn670,Leu671]-Amyloid b/A4 Precursor Protein 770 (667-676)-(2,4-dinitrophenyl)Lys-Arg-Arg Amide ³96 % (HPLC) Thimet oligopeptidase (TOP) A1347
3-Nitrotetrazolium blue chloride BioChemika ³75 % (AT) Thiophene-2-carbonyl-CoA monooxygenase 09972
Nitrotetrazolium blue chloride BioChemika ³88 % (AT) Thiophene-2-carbonyl-CoA monooxygenase 74030
Nitrotetrazolium blue chloride BioChemika ~10 mg/tablet nitrotetrazolium blue chloride Thiophene-2-carbonyl-CoA monooxygenase 74031
Nitrotetrazolium blue chloride BioChemika ³98.0 % (calc. based on dry substance, AT) Thiophene-2-carbonyl-CoA monooxygenase 74032
Tetrazolium Blue Chloride BioChemika indicator for germination, for microbiology ³90% (AT) Thiophene-2-carbonyl-CoA monooxygenase 88190
N-Benzoyl-Phe-Val-Arg-p-nitroanilide hydrochloride BioChemika ³99.0 % (TLC) Thrombin 13042
N-Benzoyl-Phe-Val-Arg 4-methoxy-b-naphthylamide hydrochloride Thrombin B1260
N-Benzoyl-Phe-Val-Arg-p-nitroanilide hydrochloride Thrombin B7632
Boc-b-benzyl-Asp-Pro-Arg-7-amido-4-methylcoumarin hydrochloride ~95% Thrombin B4028
Boc-Val-Pro-Arg-7-amido-4-methylcoumarin hydrochloride Thrombin B9385
4-Methylumbelliferyl 4-guanidinobenzoate hydrochloride hydrate crystalline Thrombin M4149
4-Methylumbelliferyl 4-guanidinobenzoate hydrochloride hydrate BioChemika for fluorescence Thrombin 51010
4-Nitrophenyl 4-guanidinobenzoate hydrochloride BioChemika ³95.0 % (AT) Thrombin 51015
Sar-Pro-Arg p-nitroanilide dihydrochloride Thrombin S9009
Thrombin generation chromogenic substrate ³90% (TLC) solid Thrombin T3068
N-p-Tosyl-Gly-Pro-Arg 7-amido-4-methylcoumarin hydrochloride Thrombin T0273
N-(p-Tosyl)-Gly-Pro-Arg p-nitroanilide acetate salt Thrombin T1637
Na-p-Tosyl-L-arginine methyl ester hydrochloride BioChemika for the determination of trypsin ³99.0 % (AT) Thrombin 90170
N-(p-Tosyl)-Gly-Pro-Arg p-nitroanilide acetate salt BioChemika ³98.0 % (TLC) Thrombin 90175
N-Acetyl-L-phenylalanyl-3,5-diiodo-L-tyrosine BioChemika ³97.0 % (CHN) Thyroid-hormone transaminase 01408
pGlu-Gly-Arg-7-amido-4-trifluoromethylcoumarin ³95 % (HPLC) lyophilized powder Tissue Plasinogen Activator P5615
Tissue plasminogen activator chromogenic substrate ³95 % (HPLC) solid Tissue Plasinogen Activator T2943
D-Val-Leu-Lys 7-amido-4-methylcoumarin ³99 % (HPLC) lyophilized powder Tissue Plasinogen Activator V3138
D-Val-Leu-Lys 7-amido-4-trifluoromethylcoumarin lyophilized powder Tissue Plasinogen Activator V3013
D-Val-Leu-Lys 4-methoxy-2-naphthylamide solid Tissue Plasinogen Activator V3263
D-Val-Phe-Lys p-Nitroanilide dihydrochloride ³97% (HPLC) Tissue Plasinogen Activator V6884
Pefachrome® tPA BioChemika ³95.0 % (HPLC) Tissue Plasminogen Activator 94174
3-Amino-2,3-dihydrobenzoic acid hydrochloride purum ³98.0% (calc. based on dry substance, T) Transaminase, gamma- 07710
AQUANAL®-plus ammonium (NH4) 0.2-8.0 mg/l refill pack for 37400 Triosephosphate isomerase 37440
Dihydroxyacetone phosphate dilithium salt BioChemika ~95 % (enzymatic) Triosephosphate isomerase 37440
Dihydroxyacetone phosphate dilithium salt technical ~90 % (enzymatic) Triosephosphate isomerase 37442
AQUANAL®-plus chromium (Cr) 0.005-0.1 mg/l refill pack for 37402 Triosephosphate isomerase 37442
Z-L-arginine 7-amido-4-methylcoumarin hydrochloride BioChemika for fluorescence ³90% (TLC) Trypsin 95938
Azo-Casein BioChemika Trypsin 11610
Azo-Casein sodium salt BioChemika lyophilized Trypsin 11615
Boric anhydride granulated Trypsin 11615
Na-Benzoyl-L-arginine ethyl ester hydrochloride Trypsin B4500
Na-Benzoyl-DL-arginine 4-nitroanilide hydrochloride ³98% Trypsin B4875
Na-Benzoyl-L-arginine ethyl ester hydrochloride BioChemika ³99.0% (AT) Trypsin 12880
Na-Benzoyl-L-arginine 4-nitroanilide hydrochloride BioChemika ³98.0 % (enzymatic) Trypsin 12915
Cobalt puriss. ³99.9% wire Trypsin 12915
Na-Benzoyl-L-arginyl-4-amino-benzoic acid hydrochloride BioChemika ³99.0 % (sum of enantiomers, TLC) Trypsin 12925
Na-Benzoyl-DL-arginine b-naphthylamide hydrochloride BioChemika ³97.0 % (AT) Trypsin 12910
Na-Benzoyl-DL-arginine 4-nitroanilide hydrochloride BioChemika ³98.0% (AT) Trypsin 12920
Na-Benzoyl-L-isoleucyl-L-glutamyl-glycyl-L-arginine-4-nitroanilide BioChemika ³97.0 % (HPLC) Trypsin 87528
N-Benzoyl-Phe-Val-Arg-p-nitroanilide hydrochloride BioChemika ³99.0 % (TLC) Trypsin 13042
N-Benzoyl-Phe-Val-Arg-p-nitroanilide hydrochloride Trypsin B7632
Boc-Gln-Ala-Arg-7-amido-4-methylcoumarin hydrochloride ~95% Trypsin B4153
Casein, Resorufin-labeled BioChemika Trypsin 22097
Z-Gly-Gly-Arg b-naphthylamide hydrochloride ~98% Trypsin C7784
Hide–Remazol Brilliant Blue R Trypsin H6268
Hide–Remazol Brilliant Blue R BioChemika Trypsin 89435
4-Methylumbelliferyl 4-guanidinobenzoate hydrochloride hydrate crystalline Trypsin M4149
4-Methylumbelliferyl 4-guanidinobenzoate hydrochloride hydrate BioChemika for fluorescence Trypsin 51010
N,N-Dimethylcasein BioChemika for spectrophotometric det. of enzyme activity Trypsin 39890
Na-Benzoyl-L-arginine-7-amido-4-methylcoumarin hydrochloride Trypsin B7260
Na-Benzoyl-L-arginine 4-methoxy-b-naphthylamide hydrochloride Trypsin B0885
Na-Benzoyl-L-arginine methyl ester bicarbonate salt Trypsin B4256
Na-Benzoyl-L-arginine 4-nitroanilide hydrochloride Trypsin B3133
Na-Benzoyl-L-arginine 4-nitroanilide hydrochloride Trypsin B3279
Na-Benzoyl-DL-arginine b-naphthylamide hydrochloride Trypsin B4750
Z-Arg-Arg-Arg 4-methoxy-b-naphthylamide Trypsin C5645
4-Nitrophenyl 4-guanidinobenzoate hydrochloride BioChemika ³95.0 % (AT) Trypsin 51015
Pefachrome® Fxa BioChemika ³95.0 % (HPLC) Trypsin 77739
Pefachrome® tPA BioChemika ³95.0 % (HPLC) Trypsin 94174
Pefachrome® Try BioChemika ³95.0 % (HPLC) Trypsin 77723
Na-p-Tosyl-L-arginine methyl ester hydrochloride BioChemika for the determination of trypsin ³99.0 % (AT) Trypsin 90170
N-Acetyl-Lys-Pro-Arg-7-amido-4-trifluoromethylcoumarin >95 % (HPLC) solid Tryptase C6608
Tyramine BioChemika for fluorescence ³99.0% (TLC) Tyramine dehydrogenase 93810
Nedd8 human ³95 % (SDS-PAGE) recombinant, expressed in Escherichia coli buffered aqueous solution Ubc12 N8529
Nedd8 Precursor human ³95 % (SDS-PAGE) recombinant, expressed in Escherichia coli buffered aqueous solution Ubc12 N2030
SUMO-1 human ³95 % (SDS-PAGE) recombinant, expressed in Escherichia coli UbcH9 S0314
SUMO-1 human ³95 % (SDS-PAGE) recombinant, expressed in Escherichia coli (GST-tagged) UbcH9 S0439
Urea BioChemika Ultra ³99.5% (T) Urea carboxylase 51458
3-(1-Acetylacetonylazo)phthalhydrazide BioChemika ³97.0 % (HPLC) Urease 17751
Urea BioChemika Ultra ³99.5% (T) Urease 51458
Na-p-Tosyl-L-arginine methyl ester hydrochloride BioChemika for the determination of trypsin ³99.0 % (AT) Urinary esterase 90170
N-Benzoyl-Val-Gly-Arg p-nitroanilide hydrochloride Urokinase B4758
Z-Gly-Gly-Arg 7-amido-4-methylcoumarin hydrochloride Urokinase C9396
Z-Gly-Gly-Arg 4-methoxy-b-naphthylamide hydrochloride Urokinase C5770
N-Glutaryl-Gly-Arg-7-amido-4-methylcoumarin hydrochloride Urokinase G2386
Gly-Arg-p-nitroanilide dihydrochloride Urokinase G8148
Pefachrome® uPA BioChemika ³95.0 % (HPLC) Urokinase 77713
pGlu-Gly-Arg-7-amido-4-trifluoromethylcoumarin ³95 % (HPLC) lyophilized powder Urokinase P5615
Iodonitrotetrazolium chloride BioChemika for histol. analysis (of oxidases) ³97.0% (AT) Xanthine dehydrogenase 58030
4-O-Methyl-D-glucurono-D-xylan dyed with Remazol brilliant blue R BioChemika 15 % as dye Xylanase, endo-1,4-beta- 66960
Ostazin Brilliant Red H-3B hydroxyethyl cellulose Xylanase, endo-1,4-beta- O6879
4-Methylumbelliferyl-b-D-xylopyranoside Xylosidase, beta- M7008
4-Methylumbelliferyl-b-D-xylopyranoside BioChemika ³99.0 % (TLC) Xylosidase, beta- 69617
4-Nitrophenyl b-D-xylopyranoside ³98% Xylosidase, beta- N2132
4-Nitrophenyl b-D-xylopyranoside BioChemika ³99.0 % (TLC) Xylosidase, beta- 73750
LAT human ~50 % (glutathione-agarose chromatography) recombinant, expressed in Escherichia coli buffered aqueous glycerol solution ZAP 70 Kinase L9413
  A to C   D to N   P to Z