Life Science > Metabolomics > Learning Center > General PolyAmino Acids FAQ
General PolyAmino Acids FAQ
  1. What is a polyamino acid?
  2. What is a homo polyamino acid?
  3. What is a random co-polyamino acid?
  4. Are the polyamino acid molecular weights average or absolute?
  5. What are some applications of polyamino acids?
What is a polyamino acid?
A polyamino acid is a synthetic polymer made up of many repeating units of amino acid(s).
[ top ]   [Main FAQ list]
What is a homo polyamino acid?
A homo polyamino acid is a polymer made up of a single amino acid as the repeating unit.
[ top ]   [Main FAQ list]
What is a random co-polyamino acid?
A random co-polyamino acid is a polymer made from two or more different amino acids that repeat in a random sequence.
[ top ]   [Main FAQ list]
Are the polyamino acid molecular weights average or absolute?
All polyamino acid molecular weights are average molecular weights and not absolute. Polydispersity is calculated from the ratio of Mw/Mn (Mw is the weight average molecular weight and Mn is the number average molecular weight).
[ top ]   [Main FAQ list]
What are some applications of polyamino acids?
Polyamino acids are used in a variety of applications.
    Clinical applications include:
  • Micro encapsulation
  • Nano and micro particles for tissue-targeted drug delivery and for the delivery of biologics
  • Vaccines for hepatitis C, multiple scleroses, emphysema
  • Gene therapy
  • Immuno suppressants
    Non-clinical applications include:
  • High throughput screening
  • Substrates for the purification of protein kinases
  • Cell adhesion
[ top ]   [Main FAQ list]