Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Metabolomics > Learning Center > Metabolic Pathways
Metabolomics
Metabolic Pathways
Metabolic Pathways Chart
The Metabolic Pathways Map contains over 500 hyperlinks to Sigma product listings. Just click on the metabolite name or the enzyme's E.C. number to access product information.
The 22nd (2003) edition of the IUBMB-Sigma-Nicholson Metabolic Pathways Chart contains updated pathways involved in ATP metabolism in the mitochondria and chloroplast. The "backbone" of the map is the Glycolytic Pathway followed by the TCA (Krebs) Cycle and the Respiratory Chain which together lead to the synthesis of ATP by ATP Synthase. ATP is the source of most of the energy required for all life. Many biosynthetic and breakdown pathways of metabolism such as carbohydrates, amino acids, lipids are associated with this backbone and are differentiated by the use of color. Human metabolism is distiguished where possible by the use of black arrows. Some 550 reactions are identified by their IUBMB Enzyme Commission (EC) numbers which are then indexed.
The 22nd edition is now supplemented on-line by Minimaps which take selected individual pathways and enlarge them to illustrate aspects of metabolism that could not previously be shown – such as compartmentalization and regulation.
A unique and exciting development is the introduction of animated metabolic pathways - "animaps". These are being converted into Flash in which form they will become freely accessible to all on the internet. Biochemistry thus becomes more clearly BIO-chemistry and Metabolism becomes more "Meaningful, Wonderful and Fun"!
View Chart (1.21 Mb PDF file) — Use Acrobat's Zoom-In Tool when viewing chart.
The 22nd edition of the comprehensive Metabolic Pathways Chart. Features glycolysis, carbohydrate, amino acid, lipid, nucleotide, steroid, vitamin metabolism and much more.
Get Adobe Acrobat ReaderThis presentation requires the Acrobat plug-in for proper viewing.
Download the reader for FREE.
Complete List of Enzyme Commission Numbers on the Metabolic Pathways Chart
This table cross references the names of all the enzymes to the corresponding E.C.#s on the Metabolic Pathway Charts.
Inborn Errors of Metabolism Chart
Contains hyperlinks to Online Mendelian Inheritence in Man (OMIM) disease database (PDF 1.6MB)
IUBMB-Nicholson Minimaps
Selected individual pathways have been enlarged to illustrate aspects of metabolism that could not previously be shown, such as compartmentation, enzymes and regulation.
IUBMB-Nicholson Electron Transport Animation
An animated representation of how the protonmotive force is generated in mitochondria to drive ATP synthase.
IUBMB-Nicholson ATP Synthase Animation
An animated representation of the mechanism of what many consider to be the most important molecule to living systems.
IUBMB-Nicholson Glycolytic Pathway Animation
An animated representation of the reactions that convert glucose into pyruvate.
IUBMB-Nicholson Citric Acid (TCA) Cycle Animation
An animated representation of how the cell harnesses the potential energy of Acetyl-CoA into the reducing power of NADH + H+ and UQH2.
IUBMB-Nicholson Pyruvate Dehydrogenase Complex Animation
An animated representation of how pyruvate from the cytosol links to Acetyl-CoA and the TCA Cycle in the Mitochondrial Matrix.
Charts Available Chart Size Product #
Metabolic Pathways Chart 33" x 50" M3907
Metabolic Pathways Chart 12.5" x 19" M3782
Inborn Errors of Metabolism Chart 21" x 30" I8014
Order Your Copy Today!
About the Author
Dr. Donald Nicolson
Dr. Donald Nicholson received his B.Sc. in 1936 and his Ph.D in 1940, both in Chemistry from London University. He then went to work at Boots Pure Drug Company at Nottingham on the development and production of the earliest sulphonamide antibacterial drugs. In 1946 he joined the department of Bacteriology at the School of Medicine in Leeds as an I.C.I Fellow in Chemotherapy and remained there as Lecturer or Senior Lecturer in Microbiology until "retirement" in 1981. Now over 90, Dr. Nicholson is still actively engaged in the creation of smaller and more detailed "minimaps" and especially "animaps". In recognition of his work he has received an Honorary D.Sc. and has been made one of only two Honorary Life Members of the IUBMB.
Learn more about Dr. Nicholson and how he created the Metabolic Pathway Charts.
Publication Data
Designed by Donald Nicholson
Prepared and published by Sigma-Aldrich Co., St.Louis, MO
©2002 International Union of Biochemistry and Molecular Biology