|
Automated Sequencing
Promospin Sale
|
|
Promo Code 989
Worldwide Customers
No Expiration Date
|
| Great Savings on SigmaSpin Sequencing Reaction Clean-Up Plates! |
Product Number |
Product Description |
Plates
|
List Price |
Discount Price |
| S4309 |
SigmaSpin™ 96-Well Post-Reaction Clean-Up Plates |
2
|
$163.20 |
$81.60 |
| S4434 |
SigmaSpin™ 96-Well Post-Reaction Clean-Up Plates |
10
|
$707.00 |
$353.50 |
| S4559 |
SigmaSpin™ 96-Well Post-Reaction Clean-Up Plates |
50
|
$2530.00 |
$1265.00 |
SigmaSpin™ 96-Well Post-Reaction Clean-Up Plates
SigmaSpin™ 96-Well Post-Reaction Clean-Up plates provide a fast, simple, and highly efficient method for removing unincorporated dyes, excess salts, and other interfering reaction components. Each plate is packed with a pre-hydrated size-exclusion resin, equilibrated with molecular biology grade water, and supplied in our unique plate design with long drip directors to minimize contamination between samples. The plate design also includes a snap-cap bottom seal and a foil seal top to ensure that the resin remains hydrated. SigmaSpin™ has been tested in high-throughput genome centers and core facilities with ABI Prism® 3700, 3100, 310 and 377. Each well can accept sample volumes up to 20 µl.
|
Flow diagram for SigmaSpin Plate
|
Ideal for removing
- Dye-terminator nucleotides and primers from sequencing reactions
- Radiolabeled nucleotides, primers, and fluorescent dyes from nucleic acid probe labeling reactions
|
SigmaSpin™ Post-Reaction Clean-Up 96-well plates come ready for immediate use.
|
Features and Benefits
- Validated with most automated sequencers and dye terminator chemistries
- Pre-qualified size-exclusion resin guarantees optimum performance
- Unique drip directors prevent cross-contamination between samples during collection
- Plates are sealed to eliminate leakage or drying during shipping or storage
- Suitable for use with multi-channel pipettes and automated workstations